1 - 7 of 7 products displayed
  • HNF1A Polyclonal Antibody for Western Blot, IF, ICC, IHC (P) and IP
  • Host Rabbit
  • Target Species Human, Mouse, Rat
  • Conjugate Unconjugated
  • Catalog # PA522310
  • Size 100µL
Applications
Immunocytochemistry (1:100-1:1000), Immunofluorescence (1:100-1:1000), Immunohistochemistry (Paraffin) (1:100-1:1000), Immunoprecipitation (Assay-Dependent), Western Blot (1:1000-1:10,000)
Alternate names:
Transcription factor 1; Liver-specific transcription factor LF-B1
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA522310 349.00
Check your price
  • HNF1A Monoclonal Antibody for Western Blot
  • Host Mouse
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # MA517267
  • Size 100µL
Applications
Western Blot (1:500-1:3000)
Alternate names:
Transcription factor 1; Liver-specific transcription factor LF-B1
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA517267 349.00
Check your price
  • HNF1A Monoclonal Antibody for Western Blot and IHC (F)
  • Host Mouse
  • Target Species Hamster, Human, Mouse, Rat, Xenopus
  • Conjugate Unconjugated
  • Catalog # MA125417
  • Size 25µg
Applications
Immunohistochemistry (Frozen) (Assay Dependent), Western Blot (0.5-2 µg/ml)
Alternate names:
Transcription factor 1; Liver-specific transcription factor LF-B1; XLFB1a; Transcription factor 1-A; LFB1
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA125417 475.00
Check your price
  • PCBD1 Polyclonal Antibody for Western Blot, IHC and Flow
  • Host Rabbit
  • Target Species Human, Mouse
  • Conjugate Unconjugated
  • Catalog # PA526121
  • Size 400µL
Applications
Flow Cytometry (1:10-1:50), Immunohistochemistry (1:50-1:100), Western Blot (1:1000)
Alternate names:
Pterin carbinolamine dehydratase; Phenylalanine hydroxylase-stimulating protein; Dimerization cofactor of hepatocyte nuclear factor 1-alpha; 4-alpha-hydroxy-tetrahydropterin dehydratase
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA526121 349.00
Check your price
  • TCF2 Polyclonal Antibody for Western Blot
  • Host Rabbit
  • Target Species Human, Mouse
  • Conjugate Unconjugated
  • Catalog # PA124459
  • Size 50µg
Applications
Western Blot (0.1-2 µg/ml)
Alternate names:
Transcription factor 2; Homeoprotein LFB3; Variant hepatic nuclear factor 1
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA124459 425.00
Check your price
  • TCF2 Polyclonal Antibody for Western Blot, ICC and pep-ELISA
  • Host Goat
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # PA518642
  • Size 100ug
Applications
Immunocytochemistry (Assay-dependent), Western Blot (0.1-0.3 ug/ml)
Alternate names:
Variant hepatic nuclear factor 1; Transcription factor 2; Homeoprotein LFB3
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA518642 349.00
Check your price
  • HNF1A Polyclonal Antibody for Western Blot, IF and ICC
  • Host Rabbit
  • Target Species Human, Mouse
  • Conjugate Unconjugated
  • Catalog # PA535356
  • Size 400µL
Applications
Immunocytochemistry (1:10-1:50), Immunofluorescence (1:10-1:50), Western Blot (1:1000)
Alternate names:
Transcription factor 1; Liver-specific transcription factor LF-B1
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA535356 349.00
Check your price