1 - 6 of 6 products displayed
  • hnRNP A1 Polyclonal Antibody for Western Blot, IF, ICC and IHC (P)
  • Host Rabbit
  • Target Species Human, Mouse, Rat
  • Conjugate Unconjugated
  • Catalog # PA528385
  • Size 100 µL
Applications
Immunocytochemistry (1:100-1:1000), Immunofluorescence (1:100-1:1000), Immunohistochemistry (Paraffin) (1:100-1:1000), Western Blot (1:500-1:3000)
Alternate names:
hnRNP core protein A1; Single-strand RNA-binding protein; Helix-destabilizing protein; Topoisomerase-inhibitor suppressed; Single-strand-binding protein; HDP-1
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA528385 349.00
Check your price
  • hnRNP A1 Polyclonal Antibody for Western Blot, IF, ICC, IHC (P) and IP
  • Host Rabbit
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # PA519431
  • Size 100 µg
Applications
Immunofluorescence (Assay-Dependent), Immunohistochemistry (Paraffin) (5 µg/ml), Immunoprecipitation (Assay-Dependent), Western Blot (1:500-1:1000)
Alternate names:
hnRNP core protein A1; Single-strand RNA-binding protein; Helix-destabilizing protein
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA519431 349.00
Check your price
  • hnRNP A1 Polyclonal Antibody for Western Blot and IHC (P)
  • Host Rabbit
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # PA527429
  • Size 100 µL
Applications
Immunohistochemistry (Paraffin) (1:100-1:1000), Western Blot (1:500-1:3000)
Alternate names:
hnRNP core protein A1; Single-strand RNA-binding protein; Helix-destabilizing protein
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA527429 349.00
Check your price
  • hnRNP A1 Monoclonal Antibody for Western Blot, IF, ICC, IP and ELISA
  • Host Mouse
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # MA126736
  • Size 50 µL
Applications
ELISA (Assay Dependent), Immunocytochemistry (Assay-dependent), Immunofluorescence (Assay Dependent), Immunoprecipitation (Assay Dependent), Western Blot (Assay Dependent)
Alternate names:
hnRNP core protein A1; Single-strand RNA-binding protein; Helix-destabilizing protein
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA126736 375.00
Check your price
  • hnRNP A1L2 Polyclonal Antibody for Western Blot and IHC (P)
  • Host Rabbit
  • Target Species Horse, Guinea pig, Human, Mouse, Pig, Rabbit, Rat, Zebrafish, Bovine, Dog
  • Conjugate Unconjugated
  • Catalog # PA541732
  • Size 100 µl
Applications
Immunohistochemistry (Paraffin) (4-8 µg/ml), Western Blot (0.2-1 µg/ml)
Alternate names:
hnRNP core protein A1-like 2
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA541732 309.00
Check your price
  • hnRNP A1L2 Polyclonal Antibody for Western Blot and IHC (P)
  • Host Rabbit
  • Target Species Bovine, Dog, Horse, Guinea pig, Human, Mouse, Rabbit, Rat
  • Conjugate Unconjugated
  • Catalog # PA541731
  • Size 100 µl
Applications
Immunohistochemistry (Paraffin) (4-8 µg/ml), Western Blot (1.25 µg/ml)
Alternate names:
hnRNP core protein A1-like 2
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA541731 309.00
Check your price