1 - 30 of 30 products displayed
  • hnRNP A2B1 Antibody
  • Host Rabbit
  • Target Species Human, Mouse, Rat
  • Conjugate Unconjugated
  • Catalog # PA519577
  • Size 100 µg
Applications
Immunocytochemistry (5 µg/ml), Immunofluorescence (5 µg/ml), Western Blot (1-5 µg/ml)
Alternate names:
DKFZp779B0244, FLJ22720, HNRNPA2, HNRNPB1, HNRPA2, HNRPA2B1, HNRPB1, RNPA2, SNRPB1, heterogeneous nuclear ribonucleoprotein B1; heterogeneous nuclear ribonucleoproteins A2/B1; hnRNP A2 / hnRNP B1; hnRNP A2/B1; nuclear ribonucleoprotein particle A2 protein
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA519577 349.00
Check your price
  • hnRNP A2B1 Antibody
  • Host Rabbit
  • Target Species Human, Mouse, Rat
  • Conjugate Unconjugated
  • Catalog # PA534939
  • Size 100 µL
Applications
Immunocytochemistry (Assay-Dependent), Immunofluorescence (Assay-Dependent), Immunohistochemistry (Paraffin) (1:100-1:1000), Immunoprecipitation (1:100-1:500), Western Blot (1:5000-1:20,000)
Alternate names:
HNRNPA2, HNRNPB1, HNRPA2, HNRPA2B1, HNRPB1, RNPA2, SNRPB1, heterogeneous nuclear ribonucleoproteins A2/B1, hnRNP A2 / hnRNP B1, nuclear ribonucleoprotein particle A2 protein
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA534939 349.00
Check your price
  • hnRNP A1 Antibody
  • Host Rabbit
  • Target Species Human, Mouse, Rat
  • Conjugate Unconjugated
  • Catalog # PA528385
  • Size 100 µL
Applications
Immunocytochemistry (1:100-1:1000), Immunofluorescence (1:100-1:1000), Immunohistochemistry (Paraffin) (1:100-1:1000), Western Blot (1:500-1:3000)
Alternate names:
hnRNP core protein A1; Single-strand RNA-binding protein; Helix-destabilizing protein; Topoisomerase-inhibitor suppressed; Single-strand-binding protein; HDP-1
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA528385 349.00
Check your price
  • hnRNP A1 Antibody
  • Host Rabbit
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # PA519431
  • Size 100 µg
Applications
Immunofluorescence (Assay-Dependent), Immunohistochemistry (Paraffin) (5 µg/ml), Immunoprecipitation (Assay-Dependent), Western Blot (1:500-1:1000)
Alternate names:
hnRNP core protein A1; Single-strand RNA-binding protein; Helix-destabilizing protein
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA519431 349.00
Check your price
  • hnRNP A1 Antibody
  • Host Rabbit
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # PA527429
  • Size 100 µL
Applications
Immunohistochemistry (Paraffin) (1:100-1:1000), Western Blot (1:500-1:3000)
Alternate names:
hnRNP core protein A1; Single-strand RNA-binding protein; Helix-destabilizing protein
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA527429 349.00
Check your price
  • hnRNP L Antibody
  • Host Rabbit
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # PA519735
  • Size 100 µg
Applications
Immunofluorescence (1 µg/ml), Western Blot (1 µg/ml)
Alternate names:
P/OKcl.14, FLJ35509, HNRPL, hnRNP-L, hnRNP L
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA519735 349.00
Check your price
  • hnRNP L Antibody
  • Host Rabbit
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # PA519599
  • Size 100 µg
Applications
Immunocytochemistry (1 µg/ml), Immunofluorescence (1 µg/ml), Immunohistochemistry (Paraffin) (1 µg/ml), Western Blot (1 µg/ml)
Alternate names:
P/OKcl.14, FLJ35509, HNRPL, hnRNP-L, hnRNP L
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA519599 349.00
Check your price
  • hnRNP R Antibody
  • Host Rabbit
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # PA528473
  • Size 100 µL
Applications
Western Blot (1:500-1:3000)
Alternate names:
HNRPR, hnRNP R, hnRNP-R
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA528473 349.00
Check your price
  • hnRNP C Antibody
  • Host Rabbit
  • Target Species Human, Mouse, Rat
  • Conjugate Unconjugated
  • Catalog # PA522280
  • Size 100 µL
Applications
Immunocytochemistry (1:100-1:1000), Immunofluorescence (1:100-1:1000), Immunohistochemistry (Paraffin) (1:100-1:1000), Western Blot (1:500-1:3000)
Alternate names:
C1, C2, HNRNP, HNRPC, SNRPC, heterogeneous nuclear ribonucleoproteins C1/C2, hnRNP C1/C2, nuclear ribonucleoprotein particle C1 protein, nuclear ribonucleoprotein particle C2 protein
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA522280 349.00
Check your price
  • Phospho-hnRNP C (Ser260) Antibody
  • Host Rabbit
  • Target Species Human, Mouse
  • Conjugate Unconjugated
  • Catalog # PA538589
  • Size 100 µg
Applications
Immunohistochemistry (Paraffin) (1:50-1:100), Western Blot (1:500-1:1000)
Alternate names:
Heterogeneous nuclear ribonucleoproteins C1/C2, hnRNP C1/C2, HNRNPC, HNRPC
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA538589 309.00
Check your price
  • hnRNP H2 Antibody
  • Host Rabbit
  • Target Species Human, Mouse, Rat
  • Conjugate Unconjugated
  • Catalog # PA536254
  • Size 100 µl
Applications
Immunohistochemistry (Paraffin) (1:50-1:200), Western Blot (1:500-1:1000)
Alternate names:
Heterogeneous nuclear ribonucleoprotein H'; FTP-3
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA536254 309.00
Check your price
  • hnRNP H2 Antibody
  • Host Rabbit
  • Target Species Human, Mouse, Rat
  • Conjugate Unconjugated
  • Catalog # PA538632
  • Size 100 µg
Applications
Western Blot (1:500-1:1000)
Alternate names:
Heterogeneous nuclear ribonucleoprotein H'; FTP-3
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA538632 309.00
Check your price
  • hnRNP H2 Antibody
  • Host Rabbit
  • Target Species Mouse, Rat
  • Conjugate Unconjugated
  • Catalog # PA535895
  • Size 100 µL
Applications
Immunohistochemistry (Paraffin) (1:100-1:1000)
Alternate names:
Heterogeneous nuclear ribonucleoprotein H'
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA535895 309.00
Check your price
  • hnRNP L Antibody (Monoclonal, 4D11)
  • Host Mouse
  • Target Species Amphibian, Human, Mouse
  • Conjugate Unconjugated
  • Catalog # MA126745
  • Size 50 µL
Applications
ELISA (Assay Dependent), Immunocytochemistry (Assay-dependent), Immunofluorescence (Assay Dependent), Immunoprecipitation (Assay Dependent), Western Blot (Assay Dependent)
Alternate names:
FLJ35509, HNRPL, P/OKcl.14, hnRNP-L
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA126745 309.00
Check your price
  • HnRNP Q Antibody (Monoclonal, I8E4)
  • Host Mouse
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # MA190980
  • Size 50 µL
Applications
ELISA (Assay-dependent), Immunocytochemistry (Assay-dependent), Immunofluorescence (Assay dependent), Immunoprecipitation (Assay dependent), Western Blot (Assay dependent)
Alternate names:
Synaptotagmin-binding, cytoplasmic RNA-interacting protein; NS1-associated protein 1; Glycine- and tyrosine-rich RNA-binding protein
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA190980 399.00
Check your price
  • hnRNP C Antibody
  • Host Rabbit
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # PA524221
  • Size 400 µL
Applications
Immunohistochemistry (1:50-1:100), Western Blot (1:1000)
Alternate names:
C1, C2, HNRNP, HNRPC, SNRPC, heterogeneous nuclear ribonucleoproteins C1/C2, hnRNP C1/C2, nuclear ribonucleoprotein particle C1 protein, nuclear ribonucleoprotein particle C2 protein
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA524221 349.00
Check your price
  • HnRNP Q Antibody
  • Host Rabbit
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # PA515009
  • Size 400 µL
Applications
Immunohistochemistry (1:10-1:50), Western Blot (1:1000)
Alternate names:
Synaptotagmin-binding, cytoplasmic RNA-interacting protein; NS1-associated protein 1; Glycine- and tyrosine-rich RNA-binding protein
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA515009 349.00
Check your price
  • hnRNP A1 Antibody (Monoclonal, 9H10)
  • Host Mouse
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # MA126736
  • Size 50 µL
Applications
ELISA (Assay Dependent), Immunocytochemistry (Assay-dependent), Immunofluorescence (Assay Dependent), Immunoprecipitation (Assay Dependent), Western Blot (Assay Dependent)
Alternate names:
hnRNP core protein A1; Single-strand RNA-binding protein; Helix-destabilizing protein
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA126736 375.00
Check your price
  • hnRNP M Antibody
  • Host Rabbit
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # PA530247
  • Size 100 µL
Applications
Immunocytochemistry (1:100-1:1000), Immunofluorescence (1:100-1:1000), Immunohistochemistry (Paraffin) (1:100-1:1000), Western Blot (1:5000-1:20,000)
Alternate names:
CEAR, HNRNPM4, HNRPM, HNRPM4, HTGR1, NAGR1, hnRNP M, CEA receptor, N-acetylglucosamine receptor 1, heterogenous nuclear ribonucleoprotein M4, hnRNA-binding protein M4
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA530247 349.00
Check your price
  • hnRNP R Antibody
  • Host Rabbit
  • Target Species Bovine, Dog, Horse, Guinea pig, Human, Mouse, Rabbit, Rat
  • Conjugate Unconjugated
  • Catalog # PA541801
  • Size 100 µl
Applications
Western Blot (0.2-1 µg/ml)
Alternate names:
Heterogeneous nuclear ribonucleoprotein R, FLJ25714, HNRPR, hnRNP-R, hnRNP R
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA541801 309.00
Check your price
  • PCBP2 Antibody
  • Host Rabbit
  • Target Species Human, Mouse
  • Conjugate Unconjugated
  • Catalog # PA522350
  • Size 100 µL
Applications
Immunocytochemistry (1:100-1:1000), Immunofluorescence (1:100-1:1000), Immunohistochemistry (Paraffin) (1:100-1:1000), Western Blot (1:500-1:3000)
Alternate names:
Heterogeneous nuclear ribonucleoprotein E2; Alpha-CP2; Putative heterogeneous nuclear ribonucleoprotein X; CTBP
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA522350 349.00
Check your price
  • PCBP1 Antibody
  • Host Goat
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # PA519409
  • Size 100 ug
Applications
Western Blot (0.01-0.03 ug/ml)
Alternate names:
Nucleic acid-binding protein SUB2.3; Heterogeneous nuclear ribonucleoprotein E1; Alpha-CP1
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA519409 349.00
Check your price
  • PCBP1 Antibody
  • Host Goat
  • Target Species Human, Rat
  • Conjugate Unconjugated
  • Catalog # PA519408
  • Size 100 ug
Applications
Western Blot (0.1-0.3 ug/ml)
Alternate names:
Nucleic acid-binding protein SUB2.3; Heterogeneous nuclear ribonucleoprotein E1; Alpha-CP1
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA519408 349.00
Check your price
  • PTBP1 Antibody
  • Host Goat
  • Target Species Bovine, Dog, Human, Mouse, Pig, Rat
  • Conjugate Unconjugated
  • Catalog # PA132024
  • Size 100 µg
Applications
ELISA (1:8000), Immunohistochemistry (Paraffin) (5-10 µg/ml), Western Blot (1-3 µg/ml)
Alternate names:
Heterogeneous nuclear ribonucleoprotein I; 57 kDa RNA-binding protein PPTB-1
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA132024 399.00
Check your price
  • hnRNP DL Antibody
  • Host Rabbit
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # PA535896
  • Size 100 µL
Applications
Immunocytochemistry (1:100-1:1000), Immunofluorescence (1:100-1:1000), Immunohistochemistry (Paraffin) (1:100-1:1000), Western Blot (1:1000-1:10,000)
Alternate names:
Protein laAUF1; JKT41-binding protein; AU-rich element RNA-binding factor
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA535896 309.00
Check your price
  • hnRNP DL Antibody
  • Host Rabbit
  • Target Species Bovine, Dog, Horse, Guinea pig, Human, Mouse, Rat, Zebrafish
  • Conjugate Unconjugated
  • Catalog # PA541799
  • Size 100 µl
Applications
Immunohistochemistry (Paraffin) (4-8 µg/ml), Western Blot (1 µg/ml)
Alternate names:
Protein laAUF1; JKT41-binding protein; AU-rich element RNA-binding factor
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA541799 309.00
Check your price
  • PTBP1 Antibody (Monoclonal, 1)
  • Host Mouse
  • Target Species Human, Mouse, Rat
  • Conjugate Unconjugated
  • Catalog # 324800
  • Size 100 µg
Applications
Immunofluorescence (Assay Dependent), Immunoprecipitation (Assay Dependent), Western Blot (1-3 ug/ml)
Alternate names:
Heterogeneous nuclear ribonucleoprotein I; 57 kDa RNA-binding protein PPTB-1
Catalog # Size Price (AUD) Your Price (AUD) Qty
324800 304.00
Check your price
  • PTBP1 Antibody (Monoclonal, 7)
  • Host Mouse
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # 325000
  • Size 100 µg
Applications
Immunofluorescence (Assay Dependent), Immunoprecipitation (Assay Dependent), Western Blot (1-3 ug/ml)
Alternate names:
Heterogeneous nuclear ribonucleoprotein I; 57 kDa RNA-binding protein PPTB-1
Catalog # Size Price (AUD) Your Price (AUD) Qty
325000 309.00
Check your price
  • hnRNP U Antibody (Monoclonal, 3G6)
  • Host Mouse
  • Target Species Human, Non-human primate
  • Conjugate Unconjugated
  • Catalog # MA124632
  • Size 50 µL
Applications
ELISA (Assay Dependent), Immunocytochemistry (Assay-dependent), Immunofluorescence (Assay Dependent), Immunoprecipitation (Assay Dependent), Western Blot (Assay Dependent)
Alternate names:
pp120; p120; Scaffold attachment factor A
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA124632 349.00
Check your price
  • hnRNP M1-M4 Antibody (Monoclonal, 1D8)
  • Host Mouse
  • Target Species Bovine, Human, Mouse, Pig, Rabbit, Rat
  • Conjugate Unconjugated
  • Catalog # MA116673
  • Size 200 µL
Applications
Immunocytochemistry (1:10-1:500), Immunofluorescence (1:10-1:500), Immunohistochemistry (Frozen) (1:10-1:500), Immunohistochemistry (Paraffin) (1:10-1:500), Immunoprecipitation (1:10-1:500), Western Blot (1:500)
Alternate names:
M4 protein
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA116673 309.00
Check your price