1 - 17 of 17 products displayed
  • hnRNP C Polyclonal Antibody for Western Blot, IF, ICC and IHC (P)
  • Host Rabbit
  • Target Species Human, Mouse, Rat
  • Conjugate Unconjugated
  • Catalog # PA522280
  • Size 100 µL
Applications
Immunocytochemistry (1:100-1:1000), Immunofluorescence (1:100-1:1000), Immunohistochemistry (Paraffin) (1:100-1:1000), Western Blot (1:500-1:3000)
Alternate names:
C1, C2, HNRNP, HNRPC, SNRPC, heterogeneous nuclear ribonucleoproteins C1/C2, hnRNP C1/C2, nuclear ribonucleoprotein particle C1 protein, nuclear ribonucleoprotein particle C2 protein
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA522280 349.00
Check your price
  • Phospho-hnRNP C (Ser260) Polyclonal Antibody for Western Blot and IHC (P)
  • Host Rabbit
  • Target Species Human, Mouse
  • Conjugate Unconjugated
  • Catalog # PA538589
  • Size 100 µg
Applications
Immunohistochemistry (Paraffin) (1:50-1:100), Western Blot (1:500-1:1000)
Alternate names:
Heterogeneous nuclear ribonucleoproteins C1/C2, hnRNP C1/C2, HNRNPC, HNRPC
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA538589 309.00
Check your price
  • hnRNP H2 Polyclonal Antibody for Western Blot and IHC (P)
  • Host Rabbit
  • Target Species Human, Mouse, Rat
  • Conjugate Unconjugated
  • Catalog # PA536254
  • Size 100 µl
Applications
Immunohistochemistry (Paraffin) (1:50-1:200), Western Blot (1:500-1:1000)
Alternate names:
Heterogeneous nuclear ribonucleoprotein H'; FTP-3
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA536254 309.00
Check your price
  • hnRNP H2 Polyclonal Antibody for Western Blot
  • Host Rabbit
  • Target Species Human, Mouse, Rat
  • Conjugate Unconjugated
  • Catalog # PA538632
  • Size 100 µg
Applications
Western Blot (1:500-1:1000)
Alternate names:
Heterogeneous nuclear ribonucleoprotein H'; FTP-3
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA538632 309.00
Check your price
  • hnRNP C Polyclonal Antibody for Western Blot and IHC (P)
  • Host Rabbit
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # PA524221
  • Size 400 µL
Applications
Immunohistochemistry (1:50-1:100), Western Blot (1:1000)
Alternate names:
C1, C2, HNRNP, HNRPC, SNRPC, heterogeneous nuclear ribonucleoproteins C1/C2, hnRNP C1/C2, nuclear ribonucleoprotein particle C1 protein, nuclear ribonucleoprotein particle C2 protein
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA524221 349.00
Check your price
  • hnRNP C1/C2 Monoclonal Antibody for Western Blot, IF, ICC, IP and ELISA
  • Host Mouse
  • Target Species Hamster, Human, Non-human primate
  • Conjugate Unconjugated
  • Catalog # MA124631
  • Size 50 µL
Applications
ELISA (Assay Dependent), Immunocytochemistry (Assay-dependent), Immunofluorescence (Assay Dependent), Immunoprecipitation (Assay Dependent), Western Blot (Assay Dependent)
Alternate names:
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA124631 309.00
Check your price
  • PRMT5 Monoclonal Antibody for Western Blot and ELISA
  • Host Mouse
  • Target Species Bovine, Dog, Hamster, Human, Mouse, Non-human primate, Rat
  • Conjugate Unconjugated
  • Catalog # MA125470
  • Size 50 µL
Applications
ELISA (Assay Dependent), Western Blot (2 µg/ml)
Alternate names:
Shk1 kinase-binding protein 1 homolog; Jak-binding protein 1; Histone-arginine N-methyltransferase PRMT5; 72 kDa ICln-binding protein
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA125470 349.00
Check your price
  • PRMT5 Polyclonal Antibody for Western Blot
  • Host Rabbit
  • Target Species Human, Mouse
  • Conjugate Unconjugated
  • Catalog # PA141390
  • Size 200 µL
Applications
Western Blot (1:500-1:1000)
Alternate names:
Shk1 kinase-binding protein 1 homolog; Jak-binding protein 1; Histone-arginine N-methyltransferase PRMT5; 72 kDa ICln-binding protein
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA141390 309.00
Check your price
  • PRMT5 Monoclonal Antibody for IF, ICC, IHC (P), Flow and IP
  • Host Mouse
  • Target Species Human, Mouse
  • Conjugate Unconjugated
  • Catalog # 730054
  • Size 100 µg
Applications
Flow Cytometry (3-5 µg x 10^6 cells), Immunocytochemistry (2µg/ml), Immunofluorescence (2µg/ml), Immunohistochemistry (Paraffin) (1:20), Immunoprecipitation (Assay Dependent)
Alternate names:
Shk1 kinase-binding protein 1 homolog; Jak-binding protein 1; Histone-arginine N-methyltransferase PRMT5; 72 kDa ICln-binding protein
Catalog # Size Price (AUD) Your Price (AUD) Qty
730054 Not Orderable Online
Check your price
  • PRMT6 Polyclonal Antibody for Western Blot
  • Host Rabbit
  • Target Species Human, Mouse
  • Conjugate Unconjugated
  • Catalog # PA141091
  • Size 200 µL
Applications
Western Blot (1:500-1:1000)
Alternate names:
Histone-arginine N-methyltransferase PRMT6; Heterogeneous nuclear ribonucleoprotein methyltransferase-like protein 6
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA141091 309.00
Check your price
  • PRMT1 Monoclonal Antibody for Western Blot, IP, ELISA and IDF
  • Host Mouse
  • Target Species Bovine, Dog, Hamster, Human, Mouse, Non-human primate, Rat
  • Conjugate Unconjugated
  • Catalog # MA125468
  • Size 50 µL
Applications
ELISA (Assay Dependent), Immunodiffusion (Assay-dependent), Immunoprecipitation (Assay Dependent), Western Blot (2 µg/ml)
Alternate names:
Histone-arginine N-methyltransferase PRMT1; Interferon receptor 1-bound protein 4
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA125468 349.00
Check your price
  • PRMT1 Polyclonal Antibody for Western Blot and IHC
  • Host Rabbit
  • Target Species Human, Mouse
  • Conjugate Unconjugated
  • Catalog # PA511112
  • Size 400 µL
Applications
Immunohistochemistry (1:10-1:50), Western Blot (1:1000)
Alternate names:
Histone-arginine N-methyltransferase PRMT1; Interferon receptor 1-bound protein 4
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA511112 349.00
Check your price
  • PRMT1 Polyclonal Antibody for Western Blot, IF, ICC and IP
  • Host Rabbit
  • Target Species Bovine, Human, Mouse, Non-human primate, Rat
  • Conjugate Unconjugated
  • Catalog # PA517299
  • Size 100 µL
Applications
Immunocytochemistry (1:100), Immunofluorescence (1:100), Immunoprecipitation (1:50), Western Blot (1:1000)
Alternate names:
Histone-arginine N-methyltransferase PRMT1; Interferon receptor 1-bound protein 4
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA517299 275.00
Check your price
  • PRMT1 Polyclonal Antibody for Western Blot
  • Host Rabbit
  • Target Species Bovine, Human, Mouse, Non-human primate, Rat
  • Conjugate Unconjugated
  • Catalog # PA517302
  • Size 100 µL
Applications
Western Blot (1:1000)
Alternate names:
Histone-arginine N-methyltransferase PRMT1; Interferon receptor 1-bound protein 4
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA517302 275.00
Check your price
  • PRMT3 Monoclonal Antibody for Western Blot, IF, ICC and ELISA
  • Host Mouse
  • Target Species Human, Mouse, Rat
  • Conjugate Unconjugated
  • Catalog # MA125469
  • Size 50 µL
Applications
ELISA (Assay Dependent), Immunocytochemistry (Assay Dependent), Immunofluorescence (Assay-dependent), Western Blot (1-2 µg/ml)
Alternate names:
Heterogeneous nuclear ribonucleoprotein methyltransferase-like protein 3
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA125469 309.00
Check your price
  • PRMT3 Polyclonal Antibody for Western Blot, IF and ICC
  • Host Rabbit
  • Target Species Human, Mouse
  • Conjugate Unconjugated
  • Catalog # PA530363
  • Size 100 µL
Applications
Immunocytochemistry (1:100-1:1000), Immunofluorescence (1:100-1:1000), Western Blot (1:500-1:3000)
Alternate names:
Heterogeneous nuclear ribonucleoprotein methyltransferase-like protein 3
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA530363 349.00
Check your price
  • PRMT8 Polyclonal Antibody for Western Blot
  • Host Rabbit
  • Target Species Bovine, Dog, Horse, Guinea pig, Human, Mouse, Rabbit, Rat, Zebrafish
  • Conjugate Unconjugated
  • Catalog # PA541710
  • Size 100 µl
Applications
Western Blot (1:1000)
Alternate names:
Heterogeneous nuclear ribonucleoprotein methyltransferase-like protein 4
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA541710 309.00
Check your price