1 - 2 of 2 products displayed
  • HOXD3 Polyclonal Antibody for Western Blot
  • Host Rabbit
  • Target Species Bovine, Dog, Horse, Guinea pig, Human, Mouse, Rabbit, Rat
  • Conjugate Unconjugated
  • Catalog # PA540461
  • Size 100µl
Applications
Western Blot (0.2-1 µg/ml)
Alternate names:
Homeobox protein MH-19; Homeobox protein Hox-4.1; Homeobox protein Hox-4A
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA540461 309.00
Check your price
  • HOXD3 Polyclonal Antibody for Western Blot and IHC
  • Host Rabbit
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # PA124444
  • Size 50µg
Applications
Immunohistochemistry (Assay-dependent), Western Blot (1-2 µg/ml)
Alternate names:
Homeobox protein Hox-4A
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA124444 425.00
Check your price