1 - 2 of 2 products displayed
  • HOXD8 Antibody
  • Host Rabbit
  • Target Species Human, Mouse, Non-human primate
  • Conjugate Unconjugated
  • Catalog # PA523476
  • Size 100 ug
Applications
Western Blot (5-10 µg/ml)
Alternate names:
Homeobox protein Hox-5.4; Homeobox protein Hox-4E; Homeobox protein Hox-4.3
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA523476 349.00
Check your price
  • HOXD9 Antibody
  • Host Rabbit
  • Target Species Bovine, Guinea pig, Human, Mouse, Rabbit, Rat
  • Conjugate Unconjugated
  • Catalog # PA540887
  • Size 100 µl
Applications
Western Blot (0.2-1 µg/ml)
Alternate names:
Homeobox protein Hox-5.2; Homeobox protein Hox-4.4; Homeobox protein Hox-4C
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA540887 309.00
Check your price