1 - 3 of 3 products displayed
  • HOXA11 Polyclonal Antibody for Western Blot
  • Host Rabbit
  • Target Species Human, Mouse, Rat
  • Conjugate Unconjugated
  • Catalog # PA141396
  • Size 100 µg
Applications
Western Blot (1-3 µg/ml)
Alternate names:
Homeobox protein Hox-1.9; Homeobox protein Hox-1I
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA141396 309.00
Check your price
  • HOXA10 Polyclonal Antibody for Western Blot and IHC (P)
  • Host Goat
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # PA537866
  • Size 100 µg
Applications
Immunohistochemistry (Paraffin) (5-10 µg/ml), Western Blot (0.2-0.6 µg/ml)
Alternate names:
PL; Homeobox protein Hox-1H; Homeobox protein Hox-1.8
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA537866 309.00
Check your price
  • HOXA10 Polyclonal Antibody for Western Blot, IF and ICC
  • Host Rabbit
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # PA523781
  • Size 400 µL
Applications
Immunocytochemistry (1:10-1:50), Immunofluorescence (1:10-1:50), Western Blot (1:1000)
Alternate names:
PL; Homeobox protein Hox-1H; Homeobox protein Hox-1.8
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA523781 349.00
Check your price