1 - 2 of 2 products displayed
  • HOXA11 Polyclonal Antibody for Western Blot
  • Host Rabbit
  • Target Species Human, Mouse, Rat
  • Conjugate Unconjugated
  • Catalog # PA141396
  • Size 100µg
Applications
Western Blot (1-3 µg/ml)
Alternate names:
Homeobox protein Hox-1.9; Homeobox protein Hox-1I
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA141396 309.00
Check your price
  • HOXA11/HOXD11 Polyclonal Antibody for Western Blot, IF and ICC
  • Host Rabbit
  • Target Species Human, Mouse
  • Conjugate Unconjugated
  • Catalog # PA539209
  • Size 100µg
Applications
Immunocytochemistry (1:100-1:500), Immunofluorescence (1:100-1:500), Western Blot (1:500-1:1000)
Alternate names:
Homeobox protein Hox-4F; Homeobox protein Hox-1.9; Homeobox protein Hox-1I; Homeobox protein Hox-5.5; Homeobox protein Hox-4.6
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA539209 309.00
Check your price