1 - 30 of 30 products displayed
  • LHX3 Polyclonal Antibody for Western Blot, IF, ICC, IHC (F), IHC (P) and IP
  • Host Rabbit
  • Target Species Amphibian, Fish, Human, Mouse, Rat
  • Conjugate Unconjugated
  • Catalog # PA129491
  • Size 50µg
Applications
Immunocytochemistry (1:100-1:250), Immunofluorescence (1:100-250), Immunohistochemistry (Frozen) (Assay Dependent), Immunohistochemistry (Paraffin) (1:125-1:500), Immunoprecipitation (1:100), Western Blot (1:1250-1:2500)
Alternate names:
Homeobox protein P-LIM; Homeobox protein LIM-3
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA129491 425.00
Check your price
  • LIM1 Polyclonal Antibody for Western Blot
  • Host Rabbit
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # PA141588
  • Size 200µL
Applications
Western Blot (1:500-1:1000)
Alternate names:
Homeobox protein Lim-1
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA141588 309.00
Check your price
  • LHX3 Polyclonal Antibody for Western Blot and IHC
  • Host Rabbit
  • Target Species Bovine, Dog, Horse, Guinea pig, Human, Mouse, Rat, Zebrafish
  • Conjugate Unconjugated
  • Catalog # PA540754
  • Size 100µl
Applications
Immunohistochemistry (Assay-Dependent), Western Blot (1.25 µg/ml)
Alternate names:
Homeobox protein P-LIM; Homeobox protein LIM-3
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA540754 309.00
Check your price
  • LIM1 Polyclonal Antibody for Western Blot and IHC
  • Host Rabbit
  • Target Species Bovine, Human, Mouse, Rat, Zebrafish
  • Conjugate Unconjugated
  • Catalog # PA546855
  • Size 100µl
Applications
Immunohistochemistry (Assay-Dependent), Western Blot (0.4 µg/ml)
Alternate names:
Homeobox protein Lim-1
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA546855 309.00
Check your price
  • LIM1 Monoclonal Antibody for Western Blot and IHC
  • Host Mouse
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # MA524042
  • Size 100µg
Applications
Immunohistochemistry (8-25 µg/ml), Western Blot (1 µg/ml)
Alternate names:
Homeobox protein Lim-1
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA524042 275.00
Check your price
  • LIM1 Polyclonal Antibody for Western Blot
  • Host Rabbit
  • Target Species Bovine, Dog, Horse, Guinea pig, Human, Mouse, Rabbit, Rat
  • Conjugate Unconjugated
  • Catalog # PA540531
  • Size 100µl
Applications
Western Blot (0.2-1 µg/ml)
Alternate names:
Homeobox protein Lim-1
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA540531 309.00
Check your price
  • LHX4 Monoclonal Antibody for Western Blot, IF, ICC and ELISA
  • Host Mouse
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # MA522832
  • Size 100µg
Applications
ELISA (Assay-Dependent), Immunocytochemistry (10 µg/ml), Immunofluorescence (10 µg/ml), Western Blot (1-5 µg/ml)
Alternate names:
CPHD4, LIM homeobox protein 4, LIM/homeobox protein Lhx4
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA522832 309.00
Check your price
  • LHX6 Monoclonal Antibody for Western Blot and ELISA
  • Host Mouse
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # MA521649
  • Size 100µg
Applications
ELISA (Assay-Dependent), Western Blot (1-5 µg/ml)
Alternate names:
LIM/homeobox protein Lhx6.1
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA521649 309.00
Check your price
  • LMX1A Polyclonal Antibody for Western Blot
  • Host Rabbit
  • Target Species Human, Mouse
  • Conjugate Unconjugated
  • Catalog # PA511474
  • Size 400µL
Applications
Western Blot (1:1000)
Alternate names:
LIM/homeobox protein LMX1A; LIM/homeobox protein 1.1
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA511474 349.00
Check your price
  • LMX1A Polyclonal Antibody for Western Blot and IHC (P)
  • Host Rabbit
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # PA124402
  • Size 50µg
Applications
Immunohistochemistry (Paraffin) (4-8 µg/ml), Western Blot (0.3 µg/ml)
Alternate names:
LIM/homeobox protein LMX1A; LIM/homeobox protein 1.1
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA124402 375.00
Check your price
  • LMX1A Polyclonal Antibody for Western Blot, IF and IHC
  • Host Rabbit
  • Target Species Human, Mouse, Rat
  • Conjugate Unconjugated
  • Catalog # PA534470
  • Size 100ug
Applications
Immunofluorescence (20 ug/mL), Immunohistochemistry (2.5 ug/ml), Western Blot (1-2 ug/mL)
Alternate names:
LIM/homeobox protein LMX1A; LIM/homeobox protein 1.1
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA534470 349.00
Check your price
  • LMX1B Polyclonal Antibody for Western Blot, IF and IHC
  • Host Rabbit
  • Target Species Human, Mouse, Rat
  • Conjugate Unconjugated
  • Catalog # PA534471
  • Size 100ug
Applications
Immunofluorescence (20 ug/mL), Immunohistochemistry (5 µg/ml), Western Blot (1-2 ug/mL)
Alternate names:
LIM/homeobox protein LMX1B; LIM/homeobox protein 1.2
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA534471 349.00
Check your price
  • LHX6 Polyclonal Antibody for Western Blot and IHC (P)
  • Host Rabbit
  • Target Species Bovine, Dog, Guinea pig, Human, Mouse, Rat
  • Conjugate Unconjugated
  • Catalog # PA540560
  • Size 100µl
Applications
Immunohistochemistry (Paraffin) (4-8 µg/ml), Western Blot (1-3 µg/ml)
Alternate names:
LIM/homeobox protein Lhx6.1
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA540560 309.00
Check your price
  • LHX6 Polyclonal Antibody for Western Blot and IHC (F)
  • Host Rabbit
  • Target Species Human, Mouse
  • Conjugate Unconjugated
  • Catalog # PA519743
  • Size 100µg
Applications
Immunohistochemistry (Frozen) (1:1000), Western Blot (1 µg/ml)
Alternate names:
LIM/homeobox protein Lhx6.1
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA519743 349.00
Check your price
  • LHX6 Polyclonal Antibody for IHC
  • Host Rabbit
  • Target Species Human, Mouse
  • Conjugate Unconjugated
  • Catalog # PA511469
  • Size 400µL
Applications
Immunohistochemistry (1:10-1:50)
Alternate names:
LIM/homeobox protein Lhx6.1
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA511469 349.00
Check your price
  • LHX5 Polyclonal Antibody for Western Blot, IF, ICC and IHC
  • Host Rabbit
  • Target Species Bovine, Dog, Horse, Guinea pig, Human, Mouse, Rabbit, Rat, Zebrafish
  • Conjugate Unconjugated
  • Catalog # PA540683
  • Size 100µl
Applications
Immunocytochemistry (1:500), Immunofluorescence (1:500), Immunohistochemistry (1:500), Western Blot (0.2-1 µg/ml)
Alternate names:
Homeobox protein LIM-5; Homeobox protein LIM-2
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA540683 309.00
Check your price
  • LHX5 Polyclonal Antibody for Western Blot and IHC (F)
  • Host Goat
  • Target Species Human, Mouse, Rat
  • Conjugate Unconjugated
  • Catalog # PA547828
  • Size 100µg
Applications
Immunohistochemistry (Frozen) (5-15 µg/ml), Western Blot (1 µg/ml)
Alternate names:
Homeobox protein LIM-2
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA547828 349.00
Check your price
  • LHX9 Polyclonal Antibody for Western Blot and IHC
  • Host Rabbit
  • Target Species Bovine, Dog, Horse, Guinea pig, Human, Mouse, Pig, Rabbit, Rat, Zebrafish
  • Conjugate Unconjugated
  • Catalog # PA541160
  • Size 100µl
Applications
Immunohistochemistry (Assay-Dependent), Western Blot (1.25 µg/ml)
Alternate names:
LIM homeobox 9
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA541160 309.00
Check your price
  • LHX4 Polyclonal Antibody for Western Blot
  • Host Rabbit
  • Target Species Bovine, Dog, Horse, Guinea pig, Goat, Human, Mouse, Rat
  • Conjugate Unconjugated
  • Catalog # PA540542
  • Size 100µl
Applications
Western Blot (0.2-1 µg/ml)
Alternate names:
LIM homeobox 4, Gsh-4, Gsh4, CPHD4
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA540542 309.00
Check your price
  • LHX8 Polyclonal Antibody for Western Blot and IP
  • Host Rabbit
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # PA528948
  • Size 100µL
Applications
Immunoprecipitation (1:100-1:500), Western Blot (1:1000-1:10,000)
Alternate names:
LHX7, LIM-homeodomain protein Lhx8, LIM/homeobox protein Lhx8
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA528948 349.00
Check your price
  • LMO4 Polyclonal Antibody for Western Blot
  • Host Chicken
  • Target Species Human, Mouse, Rat
  • Conjugate Unconjugated
  • Catalog # PA125725
  • Size 50µg
Applications
Western Blot (1:2,000)
Alternate names:
LIM domain only protein 4; Breast tumor autoantigen
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA125725 425.00
Check your price
  • ISL1 Polyclonal Antibody for Western Blot, IF, ICC and IHC (P)
  • Host Rabbit
  • Target Species Human, Rat
  • Conjugate Unconjugated
  • Catalog # PA527789
  • Size 100µL
Applications
Immunohistochemistry (Paraffin) (1:100-1:1000), Western Blot (1:500-1:3000)
Alternate names:
ISLET1, Isl-1, ISL1 transcription factor, LIM/homeodomain, insulin gene enhancer protein ISL-1, islet-1
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA527789 349.00
Check your price
  • LHX5 Monoclonal Antibody for IF, ICC and ELISA
  • Host Mouse
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # MA522476
  • Size 100µg
Applications
ELISA (Assay-Dependent), Immunocytochemistry (10 µg/ml), Immunofluorescence (10 µg/ml)
Alternate names:
LIM homeobox protein 5, LIM/homeobox protein Lhx5
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA522476 309.00
Check your price
  • LHX8 Polyclonal Antibody for Western Blot
  • Host Rabbit
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # PA531927
  • Size 100µL
Applications
Western Blot (1:500-1:3000)
Alternate names:
LHX7, LIM-homeodomain protein Lhx8, LIM/homeobox protein Lhx8
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA531927 349.00
Check your price
  • LHX2 Monoclonal Antibody for Western Blot, IF and ICC
  • Host Mouse
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # MA515834
  • Size 100µL
Applications
Immunocytochemistry (1:200-1:1000), Immunofluorescence (1/200-1/1000), Western Blot (1/500-1/2000)
Alternate names:
LH2, hLhx2, LIM HOX gene 2; LIM homeobox protein 2; LIM/homeobox protein Lhx2; homeobox protein LH-2
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA515834 349.00
Check your price
  • LMO4 Polyclonal Antibody for Western Blot, IF, ICC and Flow
  • Host Rabbit
  • Target Species Human, Mouse
  • Conjugate Unconjugated
  • Catalog # PA524248
  • Size 400µL
Applications
Flow Cytometry (1:10-1:50), Immunocytochemistry (1:10-1:50), Immunofluorescence (1:10-1:50), Western Blot (1:1000)
Alternate names:
LIM domain only protein 4; Breast tumor autoantigen
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA524248 349.00
Check your price
  • LHX5 Polyclonal Antibody for Western Blot
  • Host Rabbit
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # PA531393
  • Size 100µL
Applications
Western Blot (1:500-1:3000)
Alternate names:
LIM homeobox protein 5, LIM/homeobox protein Lhx5
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA531393 349.00
Check your price
  • LHX2 Polyclonal Antibody for Western Blot
  • Host Goat
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # PA518945
  • Size 100ug
Applications
Western Blot (1-3 ug/ml)
Alternate names:
LHX2; LIM homeobox 2; LH2; MGC138390; hLhx2; LIM HOX gene 2; LIM homeobox protein 2
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA518945 349.00
Check your price
  • LMO2 Monoclonal Antibody for Western Blot and Flow
  • Host Rabbit
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # 701156
  • Size 100µg
Applications
Western Blot (1-2 ug/ml)
Alternate names:
T-cell translocation protein 2; LIM domain only protein 2; Cysteine-rich protein TTG-2
Catalog # Size Price (AUD) Your Price (AUD) Qty
701156 309.00
Check your price
  • LMO2 Oligoclonal Antibody for Western Blot, IF and ICC
  • Host Rabbit
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # 710187
  • Size 100µg
Applications
Immunocytochemistry (1-3 ug/ml), Immunofluorescence (1-3 ug/ml), Western Blot (1-2 ug/ml)
Alternate names:
T-cell translocation protein 2; LIM domain only protein 2; Cysteine-rich protein TTG-2
Catalog # Size Price (AUD) Your Price (AUD) Qty
710187 309.00
Check your price