1 - 21 of 21 products displayed
  • HRP Monoclonal Antibody for Western Blot, ICC and ELISA
  • Host Mouse
  • Target Species Tag
  • Conjugate Unconjugated
  • Catalog # MA110371
  • Size 100 ug
Applications
ELISA (Assay Dependent), Immunocytochemistry (Assay Dependent), Western Blot (Assay Dependent)
Alternate names:
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA110371 159.00
Check your price
  • HRP Polyclonal Antibody for Western Blot and ELISA
  • Host Rabbit
  • Target Species Tag
  • Conjugate Unconjugated
  • Catalog # PA126409
  • Size 100 µL
Applications
ELISA (1:16,000-1:64,000), Western Blot (Assay Dependent)
Alternate names:
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA126409 349.00
Check your price
  • Mouse Albumin Polyclonal Antibody, HRP conjugate for Western Blot and ELISA
  • Host Goat
  • Target Species Mouse
  • Conjugate HRP
  • Catalog # PA130126
  • Size 1 mg
Applications
ELISA (1:15,000-1:50,000), Western Blot (1:1,000-1:5,000)
Alternate names:
Alb-1, Alb1
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA130126 249.00
Check your price
  • Rat Albumin Polyclonal Antibody, HRP conjugate for Western Blot and ELISA
  • Host Sheep
  • Target Species Rat
  • Conjugate HRP
  • Catalog # PA129267
  • Size 1 mg
Applications
ELISA (1:20000-1:75000), Western Blot (1:1000-1:5000)
Alternate names:
Alb1, Albza
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA129267 309.00
Check your price
  • Donkey anti-Rabbit IgG (H+L) Cross-Adsorbed Secondary Antibody, HRP conjugate for Western Blot
  • Host Donkey
  • Target Species Rabbit
  • Conjugate HRP
  • Catalog # SA1200
  • Size 250 µL
Applications
Western Blot (1:60-1:400)
Alternate names:
Catalog # Size Price (AUD) Your Price (AUD) Qty
SA1200 99.00
Check your price
  • Donkey anti-Mouse IgG (H+L) Cross-Adsorbed Secondary Antibody, HRP conjugate for Western Blot
  • Host Donkey
  • Target Species Mouse
  • Conjugate HRP
  • Catalog # SA1100
  • Size 250 µL
Applications
Western Blot (1:60-1:400)
Alternate names:
Catalog # Size Price (AUD) Your Price (AUD) Qty
SA1100 90.50
Check your price
  • HRP Monoclonal Antibody for Western Blot
  • Host Mouse
  • Target Species Tag
  • Conjugate Unconjugated
  • Catalog # MA515367
  • Size 100 µL
Applications
Western Blot (1/500-1/2000)
Alternate names:
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA515367 349.00
Check your price
  • Goat anti-Rabbit IgG Secondary Antibody, WesternDot® 625 conjugate for Western Blot
  • Host Goat
  • Target Species Rabbit
  • Conjugate WesternDot® 625
  • Catalog # W10809
  • Size 25 blots
Applications
Western Blot (1:500)
Alternate names:
Catalog # Size Price (AUD) Your Price (AUD) Qty
W10809 183.00
Check your price
  • Goat anti-Rabbit IgG Secondary Antibody, WesternDot® 585 conjugate for Western Blot
  • Host Goat
  • Target Species Rabbit
  • Conjugate WesternDot® 585
  • Catalog # W10804
  • Size 25 blots
Applications
Western Blot (1:500)
Alternate names:
Catalog # Size Price (AUD) Your Price (AUD) Qty
W10804 184.00
Check your price
  • Goat anti-Rabbit IgG Secondary Antibody, WesternDot® 655 conjugate for Western Blot
  • Host Goat
  • Target Species Rabbit
  • Conjugate WesternDot® 655
  • Catalog # W10814
  • Size 25 blots
Applications
Western Blot (1:500)
Alternate names:
Catalog # Size Price (AUD) Your Price (AUD) Qty
W10814 184.00
Check your price
  • Goat anti-Rabbit IgG Secondary Antibody, WesternDot® 800 conjugate for Western Blot
  • Host Goat
  • Target Species Rabbit
  • Conjugate WesternDot® 800
  • Catalog # W10816
  • Size 25 blots
Applications
Western Blot (1:500)
Alternate names:
Catalog # Size Price (AUD) Your Price (AUD) Qty
W10816 181.00
Check your price
  • Goat anti-Mouse IgG Secondary Antibody, WesternDot® 800 conjugate for Western Blot
  • Host Goat
  • Target Species Mouse
  • Conjugate WesternDot® 800
  • Catalog # W10815
  • Size 25 blots
Applications
Western Blot (1:500)
Alternate names:
Catalog # Size Price (AUD) Your Price (AUD) Qty
W10815 180.00
Check your price
  • Goat anti-Mouse IgG Secondary Antibody, WesternDot® 655 conjugate for Western Blot
  • Host Goat
  • Target Species Mouse
  • Conjugate WesternDot® 655
  • Catalog # W10813
  • Size 25 blots
Applications
Western Blot (1:500)
Alternate names:
Catalog # Size Price (AUD) Your Price (AUD) Qty
W10813 181.00
Check your price
  • Goat anti-Mouse IgG Secondary Antibody, WesternDot® 625 conjugate for Western Blot
  • Host Goat
  • Target Species Mouse
  • Conjugate WesternDot® 625
  • Catalog # W10808
  • Size 25 blots
Applications
Western Blot (1:500)
Alternate names:
Catalog # Size Price (AUD) Your Price (AUD) Qty
W10808 182.00
Check your price
  • Goat anti-Mouse IgG Secondary Antibody, WesternDot® 585 conjugate for Western Blot
  • Host Goat
  • Target Species Mouse
  • Conjugate WesternDot® 585
  • Catalog # W10803
  • Size 25 blots
Applications
Western Blot (1:500)
Alternate names:
Catalog # Size Price (AUD) Your Price (AUD) Qty
W10803 181.00
Check your price
  • Caspase 1 Polyclonal Antibody for Western Blot and IHC (P)
  • Host Rabbit
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # PA517570
  • Size 100 µL
Applications
Immunohistochemistry (Paraffin) (1:50), Western Blot (1:1000)
Alternate names:
p45; Interleukin-1 beta-converting enzyme; Interleukin-1 beta convertase
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA517570 275.00
Check your price
  • Macrophages / Monocytes Monoclonal Antibody for IHC, IHC (F), IHC (P) and Flow
  • Host Mouse
  • Target Species Bovine, Dog, Deer, Horse, Cat, Guinea pig, Goat, Human, Mammal, Non-human primate, Pig, Rabbit, Rat
  • Conjugate Unconjugated
  • Catalog # MA180446
  • Size 200 µg
Applications
Flow Cytometry (1/50-1/100), Immunohistochemistry (Paraffin, Frozen) (1/100-1/200)
Alternate names:
60B8AG, CAGB, CFAG, CGLB, L1AG, LIAG, MAC387, MIF, MRP14, NIF, P14
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA180446 399.00
Check your price
  • PKA alpha Monoclonal Antibody for Western Blot and ARRAY
  • Host Mouse
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # MA119732
  • Size 100 µg
Applications
Dot blot (Assay-dependent), Western Blot (0.5-1 µg/ml)
Alternate names:
MGC102831, MGC48865, PKACA
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA119732 309.00
Check your price
  • CRISPLD2 Polyclonal Antibody for Western Blot
  • Host Rabbit
  • Target Species Human, Mouse, Rat
  • Conjugate Unconjugated
  • Catalog # PA543477
  • Size 100 µl
Applications
Western Blot (1:2000)
Alternate names:
LCCL domain-containing cysteine-rich secretory protein 2; Cysteine-rich secretory protein 11; Coffeecrisp
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA543477 309.00
Check your price
  • Chicken anti-Human IgG F(ab')2 Secondary Antibody, HRP conjugate for Western Blot and ELISA
  • Host Chicken
  • Target Species Human
  • Conjugate HRP
  • Catalog # SA172045
  • Size 500 µg
Applications
ELISA (Assay Dependent), Western Blot (Assay Dependent)
Alternate names:
Catalog # Size Price (AUD) Your Price (AUD) Qty
SA172045 306.00
Check your price
  • HA Tag Polyclonal Antibody, HRP conjugate for Western Blot and ELISA
  • Host Rabbit
  • Target Species Tag
  • Conjugate HRP
  • Catalog # PA129751
  • Size 100 µL
Applications
ELISA (1:5000-1:15000), Western Blot (1:1000 -1:10000)
Alternate names:
human influenza hemagglutinin, HA-tag, HA, HA Epitope Tag
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA129751 375.00
Check your price