1 - 28 of 28 products displayed
  • HP1 alpha Polyclonal Antibody for Western Blot, IF and ICC
  • Host Rabbit
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # PA529879
  • Size 100µL
Applications
Immunocytochemistry (Assay-Dependent), Immunofluorescence (Assay-Dependent), Western Blot (1:5000-1:20,000)
Alternate names:
Heterochromatin protein 1 homolog alpha; Antigen p25
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA529879 349.00
Check your price
  • HP1 alpha Polyclonal Antibody for Western Blot, IF, ICC and IHC (P)
  • Host Rabbit
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # PA519439
  • Size 100µL
Applications
Immunocytochemistry (1:500), Immunofluorescence (Assay-Dependent), Immunohistochemistry (Paraffin) (Assay-Dependent), Western Blot (1:500-1:1000)
Alternate names:
Heterochromatin protein 1 homolog alpha; Antigen p25
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA519439 349.00
Check your price
  • Phospho-HP1 alpha (Ser92) Polyclonal Antibody for IHC (P)
  • Host Rabbit
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # PA537791
  • Size 100µl
Applications
Immunohistochemistry (Paraffin) (1:50-1:100)
Alternate names:
Heterochromatin protein 1 homolog alpha; Antigen p25
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA537791 309.00
Check your price
  • HP1 alpha Monoclonal Antibody for Western Blot, IF, ICC, IHC (P) and IP
  • Host Rabbit
  • Target Species Human, Mouse, Non-human primate, Rat
  • Conjugate Unconjugated
  • Catalog # MA514989
  • Size 100µL
Applications
Immunocytochemistry (1:100), Immunofluorescence (1:100), Immunohistochemistry (Paraffin) (1:200), Immunoprecipitation (1:25), Western Blot (1:1000)
Alternate names:
Heterochromatin protein 1 homolog alpha; Antigen p25
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA514989 275.00
Check your price
  • HP1 alpha Polyclonal Antibody for Western Blot, IF, ICC and Flow
  • Host Rabbit
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # PA116942
  • Size 100µL
Applications
Immunocytochemistry (1:500), Immunofluorescence (1:500), Western Blot (1:500-1:1000)
Alternate names:
Heterochromatin protein 1 homolog alpha; Antigen p25
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA116942 349.00
Check your price
  • HP1 alpha/beta/gamma Polyclonal Antibody for Western Blot, IF, ICC and ChIP
  • Host Rabbit
  • Target Species Human, Mouse
  • Conjugate Unconjugated
  • Catalog # PA540091
  • Size 50µg
Applications
ChIP assay (4 µg), Immunocytochemistry (1:500), Immunofluorescence (1:500), Western Blot (1:1000)
Alternate names:
Heterochromatin protein 1 homolog alpha; Antigen p25; p25beta; Modifier 1 protein; M31; Heterochromatin protein p25; Heterochromatin protein 1 homolog beta; HP1Hsbeta; Modifier 2 protein; Heterochromatin protein 1 homolog gamma; HECH
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA540091 309.00
Check your price
  • HP1 alpha Polyclonal Antibody, DyLight® 650 conjugate for Western Blot
  • Host Rabbit
  • Target Species Human
  • Conjugate DyLight® 650
  • Catalog # PA522738
  • Size 100µL
Applications
Western Blot (1:500-1:1000)
Alternate names:
Heterochromatin protein 1 homolog alpha; Antigen p25
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA522738 375.00
Check your price
  • HP1 alpha Polyclonal Antibody, DyLight® 550 conjugate for Western Blot
  • Host Rabbit
  • Target Species Human
  • Conjugate DyLight® 550
  • Catalog # PA522741
  • Size 100µL
Applications
Western Blot (1:500-1:1000)
Alternate names:
Heterochromatin protein 1 homolog alpha; Antigen p25
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA522741 375.00
Check your price
  • HP1 alpha Polyclonal Antibody, DyLight® 488 conjugate for Western Blot
  • Host Rabbit
  • Target Species Human
  • Conjugate DyLight® 488
  • Catalog # PA522739
  • Size 100µL
Applications
Western Blot (1:500-1:1000)
Alternate names:
Heterochromatin protein 1 homolog alpha; Antigen p25
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA522739 375.00
Check your price
  • HP1 alpha Polyclonal Antibody, Biotin conjugate for Western Blot
  • Host Rabbit
  • Target Species Human
  • Conjugate Biotin
  • Catalog # PA522737
  • Size 100µL
Applications
Western Blot (1:500-1:1000)
Alternate names:
Heterochromatin protein 1 homolog alpha; Antigen p25
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA522737 375.00
Check your price
  • HP1 alpha Polyclonal Antibody, HRP conjugate for Western Blot
  • Host Rabbit
  • Target Species Human
  • Conjugate HRP
  • Catalog # PA522740
  • Size 100µL
Applications
Western Blot (1:500-1:1000)
Alternate names:
Heterochromatin protein 1 homolog alpha; Antigen p25
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA522740 375.00
Check your price
  • HP1 alpha Polyclonal Antibody for Western Blot, IF, ICC, IHC (P), Flow and IP
  • Host Rabbit
  • Target Species Bovine, Human, Mouse, Non-human primate, Rat
  • Conjugate Unconjugated
  • Catalog # PA517441
  • Size 100µL
Applications
Flow Cytometry (1:100), Immunocytochemistry (1:200), Immunofluorescence (1:200), Immunohistochemistry (Paraffin) (1:200), Immunoprecipitation (1:25), Western Blot (1:1000)
Alternate names:
Heterochromatin protein 1 homolog alpha; Antigen p25
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA517441 275.00
Check your price
  • HP1 alpha Polyclonal Antibody for Western Blot
  • Host Rabbit
  • Target Species Dog, Human, Mouse, Non-human primate
  • Conjugate Unconjugated
  • Catalog # PA523357
  • Size 100ug
Applications
Western Blot (2-4 µg/ml)
Alternate names:
Heterochromatin protein 1 homolog alpha; Antigen p25
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA523357 349.00
Check your price
  • HP1 alpha Polyclonal Antibody for Western Blot
  • Host Rabbit
  • Target Species Dog, Human, Mouse, Non-human primate
  • Conjugate Unconjugated
  • Catalog # PA523358
  • Size 100ug
Applications
Western Blot (3-5 µg/ml)
Alternate names:
Heterochromatin protein 1 homolog alpha; Antigen p25
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA523358 349.00
Check your price
  • HP1 alpha Monoclonal Antibody, Biotin conjugate for Western Blot and IP
  • Host Mouse
  • Target Species Human
  • Conjugate Biotin
  • Catalog # 730046
  • Size 100µg
Applications
Immunoprecipitation (Assay Dependent)
Alternate names:
Heterochromatin protein 1 homolog alpha; Antigen p25
Catalog # Size Price (AUD) Your Price (AUD) Qty
730046 309.00
Check your price
  • HP1 alpha Monoclonal Antibody for Western Blot, IF, ICC, IHC (P), Flow and IP
  • Host Mouse
  • Target Species Human, Mouse, Rat
  • Conjugate Unconjugated
  • Catalog # 730019
  • Size 100µg
Applications
Flow Cytometry (3-5 µg x 10^6 cells), Immunocytochemistry (2µg/ml), Immunofluorescence (1:250), Immunohistochemistry (Paraffin) (1:100), Immunoprecipitation (2 ug), Western Blot (1:2000)
Alternate names:
Heterochromatin protein 1 homolog alpha; Antigen p25
Catalog # Size Price (AUD) Your Price (AUD) Qty
730019 304.00
Check your price
  • Phospho-HP1 alpha (Ser92) Polyclonal Antibody for IHC (P) and pep-ELISA
  • Host Rabbit
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # PA539772
  • Size 100µg
Applications
Immunohistochemistry (Paraffin) (1:50-1:100), peptide-ELISA (1:40,000)
Alternate names:
Heterochromatin protein 1 homolog alpha; Antigen p25
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA539772 309.00
Check your price
  • HP1 alpha Polyclonal Antibody for Western Blot and IHC
  • Host Goat
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # PA518432
  • Size 100ug
Applications
Immunohistochemistry (2-3µg/ml), Western Blot (0.03-0.1 ug/ml)
Alternate names:
Heterochromatin protein 1 homolog alpha; Antigen p25
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA518432 349.00
Check your price
  • ATRX Polyclonal Antibody for Western Blot, IF, ICC, IHC (P) and IP
  • Host Rabbit
  • Target Species Human, Mouse
  • Conjugate Unconjugated
  • Catalog # PA521348
  • Size 100µL
Applications
Immunocytochemistry (1:100-1:1000), Immunofluorescence (1:100-1:1000), Immunohistochemistry (Paraffin) (1:100-1:1000), Immunoprecipitation (Assay-Dependent), Western Blot (1:500-1:3000)
Alternate names:
X-linked nuclear protein; Heterochromatin protein 2; HP1-BP38 protein; HP1 alpha-interacting protein; ATP-dependent helicase ATRX; Znf-HX; X-linked helicase II
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA521348 349.00
Check your price
  • Phospho-HP1 gamma (Ser83) Polyclonal Antibody for Western Blot, IF, ICC and IP
  • Host Rabbit
  • Target Species Bovine, Fruit fly, Horse, Human, Mouse, Non-human primate, Rat
  • Conjugate Unconjugated
  • Catalog # PA517210
  • Size 100µL
Applications
Immunocytochemistry (1:200), Immunofluorescence (1:200), Immunoprecipitation (1:25), Western Blot (1:1000)
Alternate names:
Modifier 2 protein; Heterochromatin protein 1 homolog gamma; HECH
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA517210 309.00
Check your price
  • HP1 gamma Polyclonal Antibody for Western Blot, IF, ICC, IHC (P) and IP
  • Host Rabbit
  • Target Species Human, Mouse
  • Conjugate Unconjugated
  • Catalog # PA530954
  • Size 100µL
Applications
Immunocytochemistry (1:100-1:1000), Immunofluorescence (1:100-1:1000), Immunohistochemistry (Paraffin) (1:100-1:1000), Immunoprecipitation (1:100-1:500), Western Blot (1:5000-1:20,000)
Alternate names:
Modifier 2 protein; Heterochromatin protein 1 homolog gamma; HECH
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA530954 349.00
Check your price
  • Defensin 1/3 Polyclonal Antibody for Western Blot and IHC (P)
  • Host Goat
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # PA519228
  • Size 100µg
Applications
Immunohistochemistry (Paraffin) (4-6µg/ml), Western Blot (1-3µg/ml)
Alternate names:
HNP-1; Defensin, alpha 1; HNP-3; Defensin, alpha 3
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA519228 349.00
Check your price
  • Defensin Monoclonal Antibody for Western Blot, IHC (F), IHC (P) and Flow
  • Host Mouse
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # MA182491
  • Size 100µg
Applications
Flow Cytometry (Assay Dependent), Immunohistochemistry (Frozen) (0.1-0.5 ug/ml), Immunohistochemistry (Paraffin) (0.2-1 ug/ml), Western Blot (Assay Dependent)
Alternate names:
HNP-4; Defensin, alpha 4; HNP-1; Defensin, alpha 1; HNP-3; Defensin, alpha 3; HD5(20-94); Defensin, alpha 5
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA182491 800.00
Check your price
  • Haptoglobin Polyclonal Antibody for IDF
  • Host Sheep
  • Target Species Dog, Horse, Cat, Human, Mouse, Pig, Rabbit, Rat
  • Conjugate Unconjugated
  • Catalog # PA533158
  • Size 1mL
Applications
Immunodiffusion (Neat)
Alternate names:
Zonulin; Liver regeneration-related protein LRRG173
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA533158 225.00
Check your price
  • Haptoglobin Monoclonal Antibody for ELISA
  • Host Mouse
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # HYB1700602
  • Size 200µl
Applications
ELISA (1:32000)
Alternate names:
Zonulin
Catalog # Size Price (AUD) Your Price (AUD) Qty
HYB1700602 425.00
Check your price
  • BAHD1 Polyclonal Antibody for Western Blot, IHC and Flow
  • Host Rabbit
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # PA526166
  • Size 400µL
Applications
Flow Cytometry (1:10-1:50), Immunohistochemistry (1:50-1:100), Western Blot (1:1000)
Alternate names:
BAH domain-containing protein 1, bromo adjacent homology domain-containing 1 protein
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA526166 349.00
Check your price
  • TRIM28 Polyclonal Antibody for Western Blot
  • Host Rabbit
  • Target Species Human, Mouse, Rat
  • Conjugate Unconjugated
  • Catalog # PA534485
  • Size 100ug
Applications
Western Blot (1-2 ug/mL)
Alternate names:
Tripartite motif-containing protein 28; RING finger protein 96; Nuclear corepressor KAP-1; KRAB-interacting protein 1; KRAB-associated protein 1; E3 SUMO-protein ligase TRIM28; KRIP-1; KRAB-A-interacting protein
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA534485 349.00
Check your price
  • TRIM28 Polyclonal Antibody for Western Blot, IF, ICC and IHC (P)
  • Host Rabbit
  • Target Species Human, Mouse, Rat
  • Conjugate Unconjugated
  • Catalog # PA1852
  • Size 100µL
Applications
Immunocytochemistry (1:50-1:500), Immunofluorescence (1:50-1:500), Immunohistochemistry (Paraffin) (1:50-1:500), Western Blot (1:500)
Alternate names:
Tripartite motif-containing protein 28; RING finger protein 96; Nuclear corepressor KAP-1; KRAB-interacting protein 1; KRAB-associated protein 1; E3 SUMO-protein ligase TRIM28; KRIP-1; KRAB-A-interacting protein
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA1852 349.00
Check your price