1 - 30 of 30 products displayed
  • HPV Type 16 E7 Monoclonal Antibody for Western Blot and ELISA
  • Host Mouse
  • Target Species Virus
  • Conjugate Unconjugated
  • Catalog # MA123072
  • Size 500 µg
Applications
ELISA (Assay dependent), Western Blot (Assay dependent)
Alternate names:
HPV16, HPV-16, HpV16gp1
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA123072 309.00
Check your price
  • HPV Type 16 E7 Monoclonal Antibody for IF and Flow
  • Host Mouse
  • Target Species Virus
  • Conjugate Unconjugated
  • Catalog # MA514132
  • Size 500 µL
Applications
Flow Cytometry (Assay Dependent), Immunofluorescence (Assay Dependent)
Alternate names:
HPV16, HPV-16, HpV16gp0
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA514132 309.00
Check your price
  • HPV Monoclonal Antibody for IHC (P)
  • Host Mouse
  • Target Species Virus
  • Conjugate Unconjugated
  • Catalog # MA512446
  • Size 500 µL
Applications
Immunohistochemistry (Paraffin) (1:20)
Alternate names:
HPV
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA512446 349.00
Check your price
  • HPV Type 16/18 E6 Monoclonal Antibody for Western Blot, IHC (P) and IP
  • Host Mouse
  • Target Species Virus
  • Conjugate Unconjugated
  • Catalog # MA146057
  • Size 100 µl
Applications
Immunohistochemistry (Paraffin) (1:10-1:500), Immunoprecipitation (1:10-1:500), Western Blot (1:100-1:2000)
Alternate names:
HPV16, HPV-16, Human papillomavirus 18, HPV18, HPV-18
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA146057 375.00
Check your price
  • HPV Type 18 E7 Monoclonal Antibody for Western Blot and ELISA
  • Host Mouse
  • Target Species Virus
  • Conjugate Unconjugated
  • Catalog # MA123076
  • Size 500 µg
Applications
ELISA (Assay dependent), Western Blot (Assay dependent)
Alternate names:
HPV18, HPV-18
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA123076 349.00
Check your price
  • HPV Type 16 Monoclonal Antibody for IHC (P)
  • Host Mouse
  • Target Species Virus
  • Conjugate Unconjugated
  • Catalog # MA134821
  • Size 1 mL
Applications
Immunohistochemistry (Paraffin) (1:50-1:100)
Alternate names:
HPV16, HPV-16
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA134821 425.00
Check your price
  • TCFL5 Polyclonal Antibody for Western Blot
  • Host Goat
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # PA518588
  • Size 100 ug
Applications
Western Blot (0.3-1 ug/ml)
Alternate names:
HPV-16 E2-binding protein 1; Cha transcription factor
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA518588 349.00
Check your price
  • TCFL5 Polyclonal Antibody for Western Blot and IHC
  • Host Rabbit
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # PA124394
  • Size 50 µg
Applications
Immunohistochemistry (Assay-dependent), Western Blot (1-10 µg/ml)
Alternate names:
HPV-16 E2-binding protein 1; Cha transcription factor
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA124394 425.00
Check your price
  • HPV Type 16 E7 Monoclonal Antibody for Western Blot
  • Host Mouse
  • Target Species Virus
  • Conjugate Unconjugated
  • Catalog # MA515439
  • Size 100 µL
Applications
Western Blot (1/500-1/2000)
Alternate names:
HPV16, HPV-16, HpV16gp1
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA515439 349.00
Check your price
  • HPV Type 16 E7 Monoclonal Antibody for Western Blot and IHC
  • Host Mouse
  • Target Species Virus
  • Conjugate Unconjugated
  • Catalog # MA515822
  • Size 100 µL
Applications
Immunohistochemistry (1/200-1/1000), Western Blot (1/500-1/2000)
Alternate names:
HPV16, HPV-16, HpV16gp2
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA515822 349.00
Check your price
  • HPV Monoclonal Antibody for Western Blot and ELISA
  • Host Mouse
  • Target Species Virus
  • Conjugate Unconjugated
  • Catalog # MA17055
  • Size 1 mL
Applications
ELISA (1:20), Western Blot (1:10)
Alternate names:
HPV
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA17055 349.00
Check your price
  • HPV Monoclonal Antibody for Western Blot and ELISA
  • Host Mouse
  • Target Species Virus
  • Conjugate Unconjugated
  • Catalog # MA17054
  • Size 1 mL
Applications
ELISA (1:20), Western Blot (1:10)
Alternate names:
HPV
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA17054 309.00
Check your price
  • HPV/BPV Polyclonal Antibody for IF, IHC and ELISA
  • Host Goat
  • Target Species Virus
  • Conjugate Unconjugated
  • Catalog # PA535416
  • Size 1 mL
Applications
ELISA (Assay-Dependent), Immunofluorescence (1:500-1:4000), Immunohistochemistry (1:500-1:4000)
Alternate names:
HPV, BPV, Human papilloma virus, bovine papilloma virus
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA535416 309.00
Check your price
  • TCFL5 Polyclonal Antibody for Western Blot and IHC
  • Host Rabbit
  • Target Species Bovine, Dog, Horse, Guinea pig, Human, Mouse, Rat
  • Conjugate Unconjugated
  • Catalog # PA540384
  • Size 100 µl
Applications
Immunohistochemistry (Assay-Dependent), Western Blot (0.2-1 µg/ml)
Alternate names:
HPV-16 E2-binding protein 1; Cha transcription factor
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA540384 309.00
Check your price
  • HPV Type 16 E7 Monoclonal Antibody for Western Blot, IHC (P) and IP
  • Host Mouse
  • Target Species Virus
  • Conjugate Unconjugated
  • Catalog # 280006
  • Size 50 µg
Applications
Immunohistochemistry (Paraffin) (Assay Dependent)
Alternate names:
HPV16E7, HPV 16-E7
Catalog # Size Price (AUD) Your Price (AUD) Qty
280006 331.00
Check your price
  • HPV Type 16 Monoclonal Antibody for Western Blot and ELISA
  • Host Mouse
  • Target Species Virus
  • Conjugate Unconjugated
  • Catalog # MA123087
  • Size 250 µg
Applications
ELISA (Assay dependent), Western Blot (Assay dependent)
Alternate names:
HPV16, HPV-16
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA123087 309.00
Check your price
  • HPV Type 16 E2 Monoclonal Antibody for Western Blot and ELISA
  • Host Mouse
  • Target Species Virus
  • Conjugate Unconjugated
  • Catalog # MA190531
  • Size 100 µg
Applications
ELISA (Assay-dependent), Western Blot (Assay dependent)
Alternate names:
HPV16, HPV-16
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA190531 375.00
Check your price
  • HPV Type 16 E2 Monoclonal Antibody for Western Blot and ELISA
  • Host Mouse
  • Target Species Virus
  • Conjugate Unconjugated
  • Catalog # MA190530
  • Size 100 µg
Applications
ELISA (Assay-dependent), Western Blot (Assay dependent)
Alternate names:
HPV16, HPV-16
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA190530 375.00
Check your price
  • SUMO1 Polyclonal Antibody for Western Blot, IHC and Flow
  • Host Rabbit
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # PA511312
  • Size 400 µL
Applications
Flow Cytometry (1:10-1:50), Immunohistochemistry (1:50-1:100), Western Blot (1:1000)
Alternate names:
Ubiquitin-like protein UBL1; Ubiquitin-like protein SMT3C; Ubiquitin-homology domain protein PIC1; Sentrin; SMT3 homolog 3; GAP-modifying protein 1
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA511312 349.00
Check your price
  • Pan SUMO Polyclonal Antibody for Western Blot and IHC (P)
  • Host Rabbit
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # PA511375
  • Size 400 µL
Applications
Immunohistochemistry (1:50-1:100), Western Blot (1:1000)
Alternate names:
Ubiquitin-like protein SMT3B; Sentrin-2; SUMO-3; SMT3 homolog 2; HSMT3; Ubiquitin-like protein UBL1; Ubiquitin-like protein SMT3C; Ubiquitin-homology domain protein PIC1; Sentrin; SMT3 homolog 3; GAP-modifying protein 1; Ubiquitin-like protein SMT3A; SUMO-2; SMT3 homolog 1
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA511375 349.00
Check your price
  • E6AP Polyclonal Antibody for Western Blot, IF, ICC, IHC (P) and Flow
  • Host Rabbit
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # PA512038
  • Size 400 µL
Applications
Flow Cytometry (1:10-1:50), Immunocytochemistry (1:10-1:50), Immunofluorescence (1:10-1:50), Immunohistochemistry (1:50-1:100), Western Blot (1:1000)
Alternate names:
Renal carcinoma antigen NY-REN-54; Oncogenic protein-associated protein E6-AP; Human papillomavirus E6-associated protein; E6AP ubiquitin-protein ligase
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA512038 349.00
Check your price
  • SUMO1 Polyclonal Antibody for Western Blot, IF, ICC, IHC and Flow
  • Host Rabbit
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # PA524560
  • Size 400 µL
Applications
Flow Cytometry (1:10-1:50), Immunocytochemistry (1:10-1:50), Immunofluorescence (1:10-1:50), Immunohistochemistry (1:50-1:100), Western Blot (1:1000)
Alternate names:
Ubiquitin-like protein UBL1; Ubiquitin-like protein SMT3C; Ubiquitin-homology domain protein PIC1; Sentrin; SMT3 homolog 3; GAP-modifying protein 1
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA524560 349.00
Check your price
  • SUMO2/SUMO3 Polyclonal Antibody for Western Blot, IF, ICC and IHC (P)
  • Host Rabbit
  • Target Species Human, Rat
  • Conjugate Unconjugated
  • Catalog # PA511373
  • Size 400 µL
Applications
Immunocytochemistry (1:10-1:50), Immunofluorescence (1:10-1:50), Immunohistochemistry (1:50-1:100), Western Blot (1:1000)
Alternate names:
Ubiquitin-like protein SMT3A; Sentrin-2; SMT3 homolog 2; Ubiquitin-like protein SMT3B; SUMO-3; HSMT3; SUMO-2; SMT3 homolog 1
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA511373 349.00
Check your price
  • Bovine Papilloma Virus Type 1 E2 Monoclonal Antibody for Western Blot, IF, ICC and ELISA
  • Host Mouse
  • Target Species Virus
  • Conjugate Unconjugated
  • Catalog # MA171539
  • Size 100 ug
Applications
ELISA (1:1000-1:5000), Immunocytochemistry (1:1000), Immunofluorescence (1:1000), Western Blot (1:1000-1:5000)
Alternate names:
BPV, BPV-1, BPV1-E2, BPV Protein E2
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA171539 325.00
Check your price
  • Bovine Papilloma Virus Type 1 E2 Monoclonal Antibody for Western Blot, IF, ICC and IP
  • Host Mouse
  • Target Species Virus
  • Conjugate Unconjugated
  • Catalog # MA171532
  • Size 100 ug
Applications
Immunocytochemistry (1:1000), Immunofluorescence (1:1000), Immunoprecipitation (Assay Dependent), Western Blot (1:1000-1:5000)
Alternate names:
BPV, BPV-1, BPV1-E2, BPV Protein E2
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA171532 309.00
Check your price
  • Bovine Papilloma Virus Type 1 E2 Monoclonal Antibody for Western Blot, IF and ICC
  • Host Mouse
  • Target Species Virus
  • Conjugate Unconjugated
  • Catalog # MA171533
  • Size 100 ug
Applications
Immunocytochemistry (1:1000), Immunofluorescence (1:1000), Western Blot (1:1000-1:3000)
Alternate names:
BPV, BPV-1, BPV1-E2, BPV Protein E2
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA171533 349.00
Check your price
  • Bovine Papilloma Virus Type 1 E2 Monoclonal Antibody for IF, ICC and ELISA
  • Host Mouse
  • Target Species Virus
  • Conjugate Unconjugated
  • Catalog # MA171536
  • Size 100 ug
Applications
ELISA (0.2-1 ug/ml), Immunocytochemistry (Assay Dependent), Immunofluorescence (Assay dependent)
Alternate names:
BPV, BPV-1, BPV1-E2, BPV Protein E2
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA171536 325.00
Check your price
  • Bovine Papilloma Virus Type 1 E2 Monoclonal Antibody for IF, ICC and ELISA
  • Host Mouse
  • Target Species Virus
  • Conjugate Unconjugated
  • Catalog # MA171537
  • Size 100 ug
Applications
ELISA (0.8-2 ug/ml), Immunocytochemistry (Assay Dependent), Immunofluorescence (Assay dependent)
Alternate names:
BPV, BPV-1, BPV1-E2, BPV Protein E2
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA171537 325.00
Check your price
  • Bovine Papilloma Virus Type 1 E2 Monoclonal Antibody for IF, ICC and ELISA
  • Host Mouse
  • Target Species Virus
  • Conjugate Unconjugated
  • Catalog # MA171538
  • Size 100 ug
Applications
ELISA (0.3-1.5 ug/ml), Immunocytochemistry (Assay Dependent), Immunofluorescence (Assay dependent)
Alternate names:
BPV, BPV-1, BPV1-E2, BPV Protein E2
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA171538 325.00
Check your price
  • Bovine Papilloma Virus Type 1 E2 Monoclonal Antibody for Western Blot, IF, ICC and IP
  • Host Mouse
  • Target Species Virus
  • Conjugate Unconjugated
  • Catalog # MA171534
  • Size 100 ug
Applications
Immunocytochemistry (1:1000), Immunofluorescence (1:1000), Immunoprecipitation (Assay Dependent), Western Blot (1:500-1:2000)
Alternate names:
BPV, BPV-1, BPV1-E2, BPV Protein E2
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA171534 399.00
Check your price