1 - 30 of 30 products displayed
  • S1P1 Polyclonal Antibody for Western Blot
  • Host Rabbit
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # PA523441
  • Size 100 ug
Applications
Western Blot (4-6 µg/ml)
Alternate names:
Sphingosine 1-phosphate receptor Edg-1; Endothelial differentiation G-protein coupled receptor 1
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA523441 349.00
Check your price
  • CCR7 Monoclonal Antibody for Western Blot and Flow
  • Host Rat
  • Target Species Human, Mouse
  • Conjugate Unconjugated
  • Catalog # MA1163
  • Size 100 ug
Applications
Flow Cytometry (5ug/1X10^6 cells), Western Blot (1:250-1:1000)
Alternate names:
MIP-3 beta receptor; Epstein-Barr virus-induced G-protein coupled receptor 1; CDw197; BLR2
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA1163 309.00
Check your price
  • MCHR1 Polyclonal Antibody for Western Blot, IHC (P) and Flow
  • Host Rabbit
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # PA524182
  • Size 400 µL
Applications
Flow Cytometry (1:10-1:50), Immunohistochemistry (1:50-1:100), Western Blot (1:1000)
Alternate names:
Somatostatin receptor-like protein; SLC-1; MCH-1R; G-protein coupled receptor 24
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA524182 349.00
Check your price
  • GABBR2 Polyclonal Antibody for Western Blot and IHC
  • Host Rabbit
  • Target Species Human, Mouse
  • Conjugate Unconjugated
  • Catalog # PA523720
  • Size 400 µL
Applications
Immunohistochemistry (1:50-1:100), Western Blot (1:1000)
Alternate names:
G-protein coupled receptor 51; HG20
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA523720 349.00
Check your price
  • RGR Polyclonal Antibody for Western Blot, IHC and Flow
  • Host Rabbit
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # PA526892
  • Size 400 µL
Applications
Flow Cytometry (1:10-1:50), Immunohistochemistry (1:50-1:100), Western Blot (1:1000)
Alternate names:
RP44, RGR-opsin, RPE retinal G-protein coupled receptor, RPE-retinal G protein-coupled receptor
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA526892 349.00
Check your price
  • GRK5 Polyclonal Antibody for Western Blot and IHC
  • Host Rabbit
  • Target Species Human, Mouse
  • Conjugate Unconjugated
  • Catalog # PA513729
  • Size 400 µL
Applications
Immunohistochemistry (1:50-1:100), Western Blot (1:1000)
Alternate names:
G protein-coupled receptor kinase GRK5
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA513729 349.00
Check your price
  • GRK6 Polyclonal Antibody for Western Blot and IHC
  • Host Rabbit
  • Target Species Human, Mouse
  • Conjugate Unconjugated
  • Catalog # PA513730
  • Size 400 µL
Applications
Immunohistochemistry (1:50-1:100), Western Blot (1:1000)
Alternate names:
G protein-coupled receptor kinase GRK6
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA513730 349.00
Check your price
  • EDG4 Polyclonal Antibody for Western Blot and IHC
  • Host Rabbit
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # PA513130
  • Size 400 µL
Applications
Immunohistochemistry (1:50-1:100), Western Blot (1:1000)
Alternate names:
Lysophosphatidic acid receptor Edg-4
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA513130 349.00
Check your price
  • GPR37 Polyclonal Antibody for Western Blot and IHC
  • Host Rabbit
  • Target Species Human, Mouse
  • Conjugate Unconjugated
  • Catalog # PA513412
  • Size 400 µL
Applications
Immunohistochemistry (1:50-1:100), Western Blot (1:1000)
Alternate names:
Parkin-associated endothelin receptor-like receptor; G-protein coupled receptor 37; Endothelin B receptor-like protein 1
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA513412 349.00
Check your price
  • GPR37 Polyclonal Antibody for Western Blot and IHC (P)
  • Host Rabbit
  • Target Species Human, Mouse
  • Conjugate Unconjugated
  • Catalog # PA513413
  • Size 400 µL
Applications
Immunohistochemistry (1:50-1:100), Western Blot (1:1000)
Alternate names:
Parkin-associated endothelin receptor-like receptor; G-protein coupled receptor 37; Endothelin B receptor-like protein 1
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA513413 349.00
Check your price
  • CXCR3 Polyclonal Antibody for Western Blot, IHC and Flow
  • Host Rabbit
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # PA523679
  • Size 400 µL
Applications
Flow Cytometry (1:10-1:50), Immunohistochemistry (1:50-1:100), Western Blot (1:1000)
Alternate names:
Interferon-inducible protein 10 receptor; G protein-coupled receptor 9; CKR-L2
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA523679 349.00
Check your price
  • CXCR3 Polyclonal Antibody for Western Blot
  • Host Rabbit
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # PA519828
  • Size 100 µg
Applications
Western Blot (1 µg/ml)
Alternate names:
Interferon-inducible protein 10 receptor; G protein-coupled receptor 9; CKR-L2
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA519828 349.00
Check your price
  • GRK1 Polyclonal Antibody for Western Blot and IHC
  • Host Rabbit
  • Target Species Human, Mouse
  • Conjugate Unconjugated
  • Catalog # PA513725
  • Size 400 µL
Applications
Immunohistochemistry (1:50-1:100), Western Blot (1:1000)
Alternate names:
G protein-coupled receptor kinase 1
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA513725 349.00
Check your price
  • HRH3 Polyclonal Antibody for Western Blot and IHC
  • Host Rabbit
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # PA511701
  • Size 400 µL
Applications
Immunohistochemistry (1:10-1:50), Western Blot (1:1000)
Alternate names:
G-protein coupled receptor 97
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA511701 349.00
Check your price
  • Phospho-GIT1 (Tyr554) Polyclonal Antibody for Western Blot and DB
  • Host Rabbit
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # PA512982
  • Size 400 µL
Applications
Dot blot (1:500)
Alternate names:
GRK-interacting protein 1; G protein-coupled receptor kinase-interactor 1; Cool-associated and tyrosine-phosphorylated protein 1
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA512982 349.00
Check your price
  • GIT1 Polyclonal Antibody for Western Blot and IHC
  • Host Rabbit
  • Target Species Human, Mouse
  • Conjugate Unconjugated
  • Catalog # PA514313
  • Size 400 µL
Applications
Immunohistochemistry (1:10-1:50), Western Blot (1:1000)
Alternate names:
GRK-interacting protein 1; G protein-coupled receptor kinase-interactor 1; Cool-associated and tyrosine-phosphorylated protein 1
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA514313 349.00
Check your price
  • GPR111 Polyclonal Antibody for Western Blot
  • Host Sheep
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # PA547845
  • Size 100 µg
Applications
Western Blot (1 µg/ml)
Alternate names:
G-protein coupled receptor PGR20; G-protein coupled receptor 111
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA547845 349.00
Check your price
  • GPR17 Polyclonal Antibody for Western Blot, IHC and Flow
  • Host Rabbit
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # PA526851
  • Size 400 µL
Applications
Flow Cytometry (1:10-1:50), Immunohistochemistry (1:50-1:100), Western Blot (1:1000)
Alternate names:
R12; P2Y-like receptor; G-protein coupled receptor 17
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA526851 349.00
Check your price
  • GRK3 Polyclonal Antibody for Western Blot and IHC
  • Host Rabbit
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # PA526101
  • Size 400 µL
Applications
Immunohistochemistry (1:50-1:100), Western Blot (1:1000)
Alternate names:
G-protein-coupled receptor kinase 3
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA526101 349.00
Check your price
  • CELSR2 Polyclonal Antibody for Western Blot, IHC (P) and Flow
  • Host Goat
  • Target Species Human, Mouse
  • Conjugate Unconjugated
  • Catalog # PA547869
  • Size 100 µg
Applications
Flow Cytometry (2.5 µg/10^6 cells), Immunohistochemistry (Paraffin) (5-15 µg/ml), Western Blot (1 µg/ml)
Alternate names:
Multiple epidermal growth factor-like domains protein 3; Flamingo homolog 3; Epidermal growth factor-like protein 2; Cadherin family member 10
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA547869 349.00
Check your price
  • GPRASP1 Polyclonal Antibody for Western Blot, IHC and Flow
  • Host Rabbit
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # PA525727
  • Size 400 µL
Applications
Flow Cytometry (1:10-1:50), Immunohistochemistry (1:50-1:100), Western Blot (1:1000)
Alternate names:
GASP, GASP-1, GASP1, G-protein coupled receptor-associated sorting protein 1
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA525727 349.00
Check your price
  • alpha Synuclein Polyclonal Antibody for Western Blot and IHC (P)
  • Host Rabbit
  • Target Species Human, Mouse
  • Conjugate Unconjugated
  • Catalog # PA513397
  • Size 400 µL
Applications
Immunohistochemistry (1:50-1:100), Western Blot (1:1000)
Alternate names:
Non-A4 component of amyloid precursor; Non-A beta component of AD amyloid
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA513397 349.00
Check your price
  • GRK7 Polyclonal Antibody for Western Blot and IHC
  • Host Rabbit
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # PA514807
  • Size 400 µL
Applications
Immunohistochemistry (1:50-1:100), Western Blot (1:1000)
Alternate names:
G protein-coupled receptor kinase GRK7
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA514807 349.00
Check your price
  • CTRP1 Polyclonal Antibody for Western Blot, IHC and Flow
  • Host Rabbit
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # PA513558
  • Size 400 µL
Applications
Flow Cytometry (1:10-1:50), Immunohistochemistry (1:50-1:100), Western Blot (1:1000)
Alternate names:
G protein-coupled receptor-interacting protein
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA513558 349.00
Check your price
  • DYKDDDDK Tag Monoclonal Antibody, HRP conjugate for Western Blot
  • Host Mouse
  • Target Species Tag
  • Conjugate HRP
  • Catalog # MA191878HRP
  • Size 50 µL
Applications
Western Blot (1:500-1:2000)
Alternate names:
FLAG Tag, DYKDDDDK tag, FLAG antibody, ECS epitope tag, DDDDK epitope tag, DDDDK, DYKDDDDK epitope tag, Enterokinase Cleavage Site epitope tag, Enterokinase Cleavage Site tag, ECS tag
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA191878HRP 159.00
Check your price
  • DYKDDDDK Tag Monoclonal Antibody, Biotin conjugate for Western Blot, IF, ICC and IP
  • Host Mouse
  • Target Species Tag
  • Conjugate Biotin
  • Catalog # MA191878BTIN
  • Size 50 µL
Applications
Immunocytochemistry (1:200), Immunofluorescence (1:200), Immunoprecipitation (1:200), Western Blot (1:500-1:1000)
Alternate names:
FLAG Tag, DYKDDDDK tag, FLAG antibody, ECS epitope tag, DDDDK epitope tag, DDDDK, DYKDDDDK epitope tag, Enterokinase Cleavage Site epitope tag, Enterokinase Cleavage Site tag, ECS tag
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA191878BTIN 159.00
Check your price
  • Phosphotyrosine Monoclonal Antibody for Western Blot, IP and ELISA
  • Host Mouse
  • Target Species Chemical
  • Conjugate Unconjugated
  • Catalog # 037799
  • Size 1 mg
Applications
ELISA (1 µg/ml), Immunoprecipitation (2-5 ug), Western Blot (0.5-1 ug/ml)
Alternate names:
Tyrosine, pTyr
Catalog # Size Price (AUD) Your Price (AUD) Qty
037799 558.00
Check your price
  • Acetyl-Histone H4 (Lys12) Monoclonal Antibody for Western Blot and ELISA
  • Host Mouse
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # MA3058
  • Size 50 µl
Applications
ELISA (1:1000), Western Blot (1:1000)
Alternate names:
H4FA, H4K12ac, H4K12a
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA3058 309.00
Check your price
  • GAP43 Polyclonal Antibody, HRP conjugate for Western Blot, IHC, IHC (F) and IHC (P)
  • Host Rabbit
  • Target Species Dog, Chicken, Human, Mouse, Non-human primate, Pig, Rat
  • Conjugate HRP
  • Catalog # PA522898
  • Size 100 µL
Applications
Immunocytochemistry (1-2 µg/ml), Immunofluorescence (1-2 µg/ml), Immunohistochemistry (Frozen) (1-2 µg/ml), Immunohistochemistry (Paraffin) (1-2 µg/ml), Western Blot (0.1-1 µg/ml)
Alternate names:
pp46; Neural phosphoprotein B-50; Growth-associated protein 43; Axonal membrane protein GAP-43; Protein F1; Calmodulin-binding protein P-57
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA522898 375.00
Check your price
  • STAT5 beta Polyclonal Antibody for Western Blot, IHC (P), IP, ELISA and GS
  • Host Rabbit
  • Target Species Human, Mouse, Rat
  • Conjugate Unconjugated
  • Catalog # 712500
  • Size 100 µg
Applications
ELISA (Assay Dependent), Gel Shift (Assay Dependent), Immunohistochemistry (Paraffin) (1:10-1:100), Immunoprecipitation (Assay Dependent), Western Blot (Assay Dependent)
Alternate names:
Signal transducer and activator of transcription 5B, STA5B, STAT5, STAT5B, transcription factor STAT5B
Catalog # Size Price (AUD) Your Price (AUD) Qty
712500 309.00
Check your price