1 - 30 of 30 products displayed
  • Phosphotyrosine Polyclonal Antibody, HRP conjugate for Western Blot and ELISA
  • Host Rabbit
  • Target Species Chemical
  • Conjugate HRP
  • Catalog # 615820
  • Size 100 µg
Applications
ELISA (1:2000), Western Blot (1:1000-2000)
Alternate names:
Tyrosine, pTyr
Catalog # Size Price (AUD) Your Price (AUD) Qty
615820 303.00
Check your price
  • Mouse Albumin Polyclonal Antibody, HRP conjugate for Western Blot and ELISA
  • Host Goat
  • Target Species Mouse
  • Conjugate HRP
  • Catalog # PA130126
  • Size 1 mg
Applications
ELISA (1:15,000-1:50,000), Western Blot (1:1,000-1:5,000)
Alternate names:
Alb-1, Alb1
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA130126 249.00
Check your price
  • Rat Albumin Polyclonal Antibody, HRP conjugate for Western Blot and ELISA
  • Host Sheep
  • Target Species Rat
  • Conjugate HRP
  • Catalog # PA129267
  • Size 1 mg
Applications
ELISA (1:20000-1:75000), Western Blot (1:1000-1:5000)
Alternate names:
Alb1, Albza
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA129267 309.00
Check your price
  • Ovalbumin Polyclonal Antibody, HRP conjugate for Western Blot and ELISA
  • Host Rabbit
  • Target Species Chicken
  • Conjugate HRP
  • Catalog # PA1196HRP
  • Size 100 µg
Applications
ELISA (1:1000-1:5000), Western Blot (1:1000-1:5000)
Alternate names:
Plakalbumin; Egg albumin; Allergen Gal d II
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA1196HRP 225.00
Check your price
  • Goat anti-Mouse IgG (H+L) Poly-HRP Secondary Antibody, HRP conjugate for Western Blot, IHC and ELISA
  • Host Goat
  • Target Species Mouse
  • Conjugate HRP
  • Catalog # 32230
  • Size 500 µL
Applications
ELISA (1:2500-1:10,000), Immunohistochemistry (1:50-1:500), Western Blot (1:2500-1:10,000)
Alternate names:
Catalog # Size Price (AUD) Your Price (AUD) Qty
32230 199.00
Check your price
  • Goat anti-Rabbit IgG (H+L) Poly-HRP Secondary Antibody, HRP conjugate for Western Blot, IHC and ELISA
  • Host Goat
  • Target Species Rabbit
  • Conjugate HRP
  • Catalog # 32260
  • Size 500 µL
Applications
ELISA (1:60-1:500), Immunohistochemistry (1:6-1:60), Western Blot (1:60-1:500)
Alternate names:
Catalog # Size Price (AUD) Your Price (AUD) Qty
32260 199.00
Check your price
  • Rat anti-Mouse IgG (Kappa light chain) Secondary Antibody, HRP conjugate for Western Blot, IHC (F) and ELISA
  • Host Rat
  • Target Species Mouse
  • Conjugate HRP
  • Catalog # MA134732
  • Size 250 µg
Applications
ELISA (0.5-1 µg/ml), Immunohistochemistry (Frozen) (Assay Dependent)
Alternate names:
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA134732 472.00
Check your price
  • Rabbit anti-Mouse IgG (H+L) Cross-Adsorbed Secondary Antibody, HRP conjugate for Western Blot, ICC, IHC and ELISA
  • Host Rabbit
  • Target Species Mouse
  • Conjugate HRP
  • Catalog # 31452
  • Size 1 mL
Applications
Flow Cytometry (1:500-1:5,000), Immunocytochemistry (1:5,000-1:100,000), Immunofluorescence (1:5,000-1:100,000), Immunohistochemistry (1:5,000-1:100,000), Immunoprecipitation (1:500-1:5,000), Western Blot (1:5,000-1:200,000)
Alternate names:
Catalog # Size Price (AUD) Your Price (AUD) Qty
31452 179.00
Check your price
  • F(ab')2-Rabbit anti-Rat IgG (H+L) Secondary Antibody, HRP conjugate for Western Blot, IHC, ELISA and IM
  • Host Rabbit
  • Target Species Rat
  • Conjugate HRP
  • Catalog # PA129927
  • Size 250 µg
Applications
ELISA (1:3000-1:12,000), Immunohistochemistry (Assay dependent), Immunomicroscopy (Assay-dependent), Western Blot (Assay dependent)
Alternate names:
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA129927 309.00
Check your price
  • Chicken anti-Mouse IgG Fab Secondary Antibody, HRP conjugate for Western Blot and ELISA
  • Host Chicken
  • Target Species Mouse
  • Conjugate HRP
  • Catalog # SA172025
  • Size 500 µg
Applications
ELISA (Assay Dependent), Western Blot (Assay Dependent)
Alternate names:
Catalog # Size Price (AUD) Your Price (AUD) Qty
SA172025 299.00
Check your price
  • Chicken anti-Mouse IgG Fc Secondary Antibody, HRP conjugate for Western Blot and ELISA
  • Host Chicken
  • Target Species Mouse
  • Conjugate HRP
  • Catalog # SA172029
  • Size 500 µg
Applications
ELISA (Assay Dependent), Western Blot (Assay Dependent)
Alternate names:
Catalog # Size Price (AUD) Your Price (AUD) Qty
SA172029 249.00
Check your price
  • Chicken anti-Mouse IgG Secondary Antibody, HRP conjugate for Western Blot and ELISA
  • Host Chicken
  • Target Species Mouse
  • Conjugate HRP
  • Catalog # SA172021
  • Size 500 µg
Applications
ELISA (1:5000)
Alternate names:
Catalog # Size Price (AUD) Your Price (AUD) Qty
SA172021 149.00
Check your price
  • Rabbit anti-Mouse IgG Secondary Antibody, HRP conjugate for Western Blot, IHC (F), IHC (P) and ELISA
  • Host Rabbit
  • Target Species Mouse
  • Conjugate HRP
  • Catalog # PA128568
  • Size 1 mg
Applications
Dot blot (Assay dependent), ELISA (1:6000-1:30,000), Immunohistochemistry (Frozen) (1:1500-1:7500), Immunohistochemistry (Paraffin) (1:1500-1:7500), Immunomicroscopy (Assay dependent), Western Blot (1:3000-1:15,000)
Alternate names:
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA128568 208.00
Check your price
  • Rabbit anti-Mouse IgG Secondary Antibody, HRP conjugate for Western Blot, IHC (F), IHC (P), ELISA, DB and IM
  • Host Rabbit
  • Target Species Mouse
  • Conjugate HRP
  • Catalog # PA128761
  • Size 100 µg
Applications
Dot blot (Assay dependent), ELISA (1:5000-1:20,000), Immunohistochemistry (Frozen) (1:200-1:1000), Immunohistochemistry (Paraffin) (1:200-1:1000), Immunomicroscopy (Assay dependent), Western Blot (1:500-1:2500)
Alternate names:
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA128761 139.00
Check your price
  • Chicken anti-Rat IgG Secondary Antibody, HRP conjugate for Western Blot, IHC (F), IHC (P), ELISA and IM
  • Host Chicken
  • Target Species Rat
  • Conjugate HRP
  • Catalog # PA130362
  • Size 1 mg
Applications
ELISA (Assay dependent), Immunohistochemistry (Frozen) (Assay dependent), Immunohistochemistry (Paraffin) (Assay dependent), Immunomicroscopy (Assay-dependent), Western Blot (Assay dependent)
Alternate names:
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA130362 379.00
Check your price
  • Rabbit anti-Rat IgG Secondary Antibody, HRP conjugate for Western Blot, IHC (F), IHC (P) and ELISA
  • Host Rabbit
  • Target Species Rat
  • Conjugate HRP
  • Catalog # PA128573
  • Size 1 mg
Applications
ELISA (1:15,000-1:65,000), Immunohistochemistry (Paraffin, Frozen) (1:500-1:2500), Immunomicroscopy (Assay dependent), Western Blot (1:1000-1:5000)
Alternate names:
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA128573 194.00
Check your price
  • Sheep anti-Rat IgG Secondary Antibody, HRP conjugate for Western Blot, IHC (F), IHC (P), ELISA and IM
  • Host Sheep
  • Target Species Rat
  • Conjugate HRP
  • Catalog # PA128642
  • Size 1 mg
Applications
ELISA (Assay dependent), Immunohistochemistry (Frozen) (Assay dependent), Immunohistochemistry (Paraffin) (Assay dependent), Immunomicroscopy (Assay-dependent), Western Blot (Assay dependent)
Alternate names:
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA128642 195.00
Check your price
  • F(ab')2-Goat anti-Rabbit IgG (H+L) Secondary Antibody, HRP conjugate for Western Blot, IHC and ELISA
  • Host Goat
  • Target Species Rabbit
  • Conjugate HRP
  • Catalog # A24531
  • Size 500 µg
Applications
ELISA (1:500-1:20,000), Immunohistochemistry (1:500-1:20,000), Western Blot (1:500-1:20,000)
Alternate names:
Catalog # Size Price (AUD) Your Price (AUD) Qty
A24531 109.00
Check your price
  • Chicken anti-Rabbit IgG (H+L) Secondary Antibody, HRP conjugate for Western Blot, IHC and ELISA
  • Host Chicken
  • Target Species Rabbit
  • Conjugate HRP
  • Catalog # A15987
  • Size 1 mg
Applications
ELISA (1:500-1:20,000), Immunohistochemistry (1:500-1:20,000), Western Blot (1:500-1:20,000)
Alternate names:
Catalog # Size Price (AUD) Your Price (AUD) Qty
A15987 179.00
Check your price
  • Donkey anti-Rabbit IgG (H+L) Secondary Antibody, HRP conjugate for Western Blot, IHC and ELISA
  • Host Donkey
  • Target Species Rabbit
  • Conjugate HRP
  • Catalog # A16023
  • Size 1 mg
Applications
ELISA (1:500-1:20,000), Immunohistochemistry (1:500-1:20,000), Western Blot (1:500-1:20,000)
Alternate names:
Catalog # Size Price (AUD) Your Price (AUD) Qty
A16023 109.00
Check your price
  • Sheep anti-Rabbit IgG (H+L) Secondary Antibody, HRP conjugate for Western Blot, IHC and ELISA
  • Host Sheep
  • Target Species Rabbit
  • Conjugate HRP
  • Catalog # A16172
  • Size 1 mg
Applications
ELISA (1:500-1:20,000), Immunohistochemistry (1:500-1:20,000), Western Blot (1:500-1:20,000)
Alternate names:
Catalog # Size Price (AUD) Your Price (AUD) Qty
A16172 109.00
Check your price
  • Goat anti-Rabbit IgG (H+L) Secondary Antibody, HRP conjugate for Western Blot, IHC and ELISA
  • Host Goat
  • Target Species Rabbit
  • Conjugate HRP
  • Catalog # A16096
  • Size 1 mg
Applications
ELISA (1:500-1:20,000), Immunohistochemistry (1:500-1:20,000), Western Blot (1:500-1:20,000)
Alternate names:
Catalog # Size Price (AUD) Your Price (AUD) Qty
A16096 109.00
Check your price
  • F(ab')2-Donkey anti-Mouse IgG (H+L) Secondary Antibody, HRP conjugate for Western Blot, IHC and ELISA
  • Host Donkey
  • Target Species Mouse
  • Conjugate HRP
  • Catalog # A24500
  • Size 500 µg
Applications
ELISA (1:500-1:20,000), Immunohistochemistry (1:500-1:20,000), Western Blot (1:500-1:20,000)
Alternate names:
Catalog # Size Price (AUD) Your Price (AUD) Qty
A24500 149.00
Check your price
  • F(ab')2-Goat anti-Mouse IgG (H+L) Secondary Antibody, HRP conjugate for Western Blot, IHC and ELISA
  • Host Goat
  • Target Species Mouse
  • Conjugate HRP
  • Catalog # A24512
  • Size 500 µg
Applications
ELISA (1:500-1:20,000), Immunohistochemistry (1:500-1:20,000), Western Blot (1:500-1:20,000)
Alternate names:
Catalog # Size Price (AUD) Your Price (AUD) Qty
A24512 109.00
Check your price
  • Chicken anti-Mouse IgG (H+L) Secondary Antibody, HRP conjugate for Western Blot, IHC and ELISA
  • Host Chicken
  • Target Species Mouse
  • Conjugate HRP
  • Catalog # A15975
  • Size 1 mg
Applications
ELISA (1:500-1:20,000), Immunohistochemistry (1:500-1:20,000), Western Blot (1:500-1:20,000)
Alternate names:
Catalog # Size Price (AUD) Your Price (AUD) Qty
A15975 179.00
Check your price
  • Donkey anti-Mouse IgG (H+L) Secondary Antibody, HRP conjugate for Western Blot, IHC and ELISA
  • Host Donkey
  • Target Species Mouse
  • Conjugate HRP
  • Catalog # A16011
  • Size 1 mg
Applications
ELISA (1:500-1:20,000), Immunohistochemistry (1:500-1:20,000), Western Blot (1:500-1:20,000)
Alternate names:
Catalog # Size Price (AUD) Your Price (AUD) Qty
A16011 109.00
Check your price
  • Rabbit anti-Mouse IgG (H+L) Secondary Antibody, HRP conjugate for Western Blot, IHC and ELISA
  • Host Rabbit
  • Target Species Mouse
  • Conjugate HRP
  • Catalog # A16160
  • Size 1 mg
Applications
ELISA (1:500-1:20,000), Immunohistochemistry (1:500-1:20,000), Western Blot (1:500-1:20,000)
Alternate names:
Catalog # Size Price (AUD) Your Price (AUD) Qty
A16160 109.00
Check your price
  • Goat anti-Mouse IgG (H+L) Secondary Antibody, HRP conjugate for Western Blot, IHC and ELISA
  • Host Goat
  • Target Species Mouse
  • Conjugate HRP
  • Catalog # A16066
  • Size 1 mg
Applications
ELISA (1:500-1:20,000), Immunohistochemistry (1:500-1:20,000), Western Blot (1:500-1:20,000)
Alternate names:
Catalog # Size Price (AUD) Your Price (AUD) Qty
A16066 109.00
Check your price
  • Chicken anti-Rat IgG (H+L) Secondary Antibody, HRP conjugate for Western Blot, IHC and ELISA
  • Host Chicken
  • Target Species Rat
  • Conjugate HRP
  • Catalog # A18727
  • Size 1 mg
Applications
ELISA (1:500-1:20,000), Immunohistochemistry (1:500-1:20,000), Western Blot (1:500-1:20,000)
Alternate names:
Catalog # Size Price (AUD) Your Price (AUD) Qty
A18727 179.00
Check your price
  • Donkey anti-Rat IgG (H+L) Secondary Antibody, HRP conjugate for Western Blot, IHC and ELISA
  • Host Donkey
  • Target Species Rat
  • Conjugate HRP
  • Catalog # A18739
  • Size 1 mg
Applications
ELISA (1:500-1:20,000), Immunohistochemistry (1:500-1:20,000), Western Blot (1:500-1:20,000)
Alternate names:
Catalog # Size Price (AUD) Your Price (AUD) Qty
A18739 109.00
Check your price