1 - 21 of 21 products displayed
  • Phospho-HSP27 (Ser78, Ser82) Polyclonal Antibody for IHC (P)
  • Host Rabbit
  • Target Species Human, Mouse, Rat
  • Conjugate Unconjugated
  • Catalog # PA536912
  • Size 100 µl
Applications
Immunohistochemistry (Paraffin) (1:50-1:200)
Alternate names:
Stress-responsive protein 27; Heat shock 27 kDa protein; Estrogen-regulated 24 kDa protein; 28 kDa heat shock protein; p25; Heat shock 25 kDa protein; Growth-related 25 kDa protein
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA536912 349.00
Check your price
  • HSP27 Polyclonal Antibody for Western Blot, IF, ICC and IHC
  • Host Rabbit
  • Target Species Human, Non-human primate, Rat
  • Conjugate Unconjugated
  • Catalog # PA1017
  • Size 100 µg
Applications
Immunocytochemistry (Assay dependent), Immunofluorescence (1:50), Immunohistochemistry (4 µg/ml), Western Blot (1:1000-1:2000)
Alternate names:
Stress-responsive protein 27; Heat shock 27 kDa protein; Estrogen-regulated 24 kDa protein; 28 kDa heat shock protein; p25; Heat shock 25 kDa protein; Growth-related 25 kDa protein
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA1017 375.00
Check your price
  • HSP27 Polyclonal Antibody for Western Blot, IF, ICC, IP and ELISA
  • Host Rabbit
  • Target Species Human, Non-human primate
  • Conjugate Unconjugated
  • Catalog # PA126358
  • Size 50 µg
Applications
ELISA (1 µg/ml), Immunocytochemistry (10-15 µg/ml), Immunofluorescence (10-15 ug/ml), Immunoprecipitation (5-10 µg/ml), Western Blot (1-2 µg/ml)
Alternate names:
Stress-responsive protein 27; Heat shock 27 kDa protein; Estrogen-regulated 24 kDa protein; 28 kDa heat shock protein
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA126358 349.00
Check your price
  • HSP27 Monoclonal Antibody for Western Blot, IF, ICC, IHC (P), IP, ELISA and IM
  • Host Mouse
  • Target Species Dog, Human, Non-human primate, Rat
  • Conjugate Unconjugated
  • Catalog # MA3015
  • Size 100 µL
Applications
ELISA (Assay dependent), Immunocytochemistry (1:500), Immunofluorescence (1:50-1:500), Immunohistochemistry (Paraffin) (1:500), Immunomicroscopy (Assay Dependent), Immunoprecipitation (2 µg), Western Blot (1:500-1:1000)
Alternate names:
Stress-responsive protein 27; Heat shock 27 kDa protein; Estrogen-regulated 24 kDa protein; 28 kDa heat shock protein; p25; Heat shock 25 kDa protein; Growth-related 25 kDa protein
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA3015 375.00
Check your price
  • Phospho-HSP27 (Ser78) Polyclonal Antibody for Western Blot
  • Host Rabbit
  • Target Species Dog, Horse, Human, Non-human primate
  • Conjugate Unconjugated
  • Catalog # PA523340
  • Size 20 ug
Applications
Western Blot (0.01-0.1 µg/ml)
Alternate names:
Stress-responsive protein 27; Heat shock 27 kDa protein; Estrogen-regulated 24 kDa protein; 28 kDa heat shock protein
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA523340 349.00
Check your price
  • HSP27 / Heat Shock Protein 27 Synthetic Peptide for Ctrl
  • Host N/A
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # PEP187
  • Size 50 µg
Applications
Control (Assay Dependent)
Alternate names:
Heat shock protein beta-1; HspB1; HSP25; Heat shock 27 kDa protein; HSP 27; Stress-responsive protein 27; SRP27; Estrogen-regulated 24 kDa protein; 28 kDa heat shock protein; HSPB1; HSP27
Catalog # Size Price (AUD) Your Price (AUD) Qty
PEP187 99.75
Check your price
  • HSP27 / Heat Shock Protein 27 Synthetic Peptide for Ctrl
  • Host N/A
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # PEP240
  • Size 50 µg
Applications
Control (Assay Dependent)
Alternate names:
Heat shock protein beta-1; HspB1; HSP25; Heat shock 27 kDa protein; HSP 27; Stress-responsive protein 27; SRP27; Estrogen-regulated 24 kDa protein; 28 kDa heat shock protein; HSPB1; HSP27
Catalog # Size Price (AUD) Your Price (AUD) Qty
PEP240 109.00
Check your price
  • HSP27 Polyclonal Antibody for Western Blot and IHC
  • Host Rabbit
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # PA514023
  • Size 400 µL
Applications
Immunohistochemistry (1:50-1:100), Western Blot (1:1000)
Alternate names:
Stress-responsive protein 27; Heat shock 27 kDa protein; Estrogen-regulated 24 kDa protein; 28 kDa heat shock protein
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA514023 349.00
Check your price
  • Phospho-HSP27 (Ser85) Polyclonal Antibody for Western Blot, IF, ICC, IHC (P), Flow and IP
  • Host Rabbit
  • Target Species Human, Rat
  • Conjugate Unconjugated
  • Catalog # PA1005
  • Size 100 µg
Applications
Flow Cytometry (3-5 µg x 10^6 cells), Immunocytochemistry (15 -25 µg/ml), Immunofluorescence (15-25 µg/ml), Immunoprecipitation (3 µg), Western Blot (0.25-2 µg/ml)
Alternate names:
Stress-responsive protein 27; Heat shock 27 kDa protein; Estrogen-regulated 24 kDa protein; 28 kDa heat shock protein
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA1005 349.00
Check your price
  • HSP27 Monoclonal Antibody, Biotin conjugate for Western Blot and IHC (P)
  • Host Mouse
  • Target Species Human, Non-human primate
  • Conjugate Biotin
  • Catalog # MA511213
  • Size 500 µL
Applications
Immunohistochemistry (Paraffin) (1-2 µg/ml), Western Blot (1-2 µg/ml)
Alternate names:
Stress-responsive protein 27; Heat shock 27 kDa protein; Estrogen-regulated 24 kDa protein; 28 kDa heat shock protein
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA511213 375.00
Check your price
  • Phospho-HSP27 (Ser15) Polyclonal Antibody for Western Blot, IF, ICC and IP
  • Host Rabbit
  • Target Species Human, Rat
  • Conjugate Unconjugated
  • Catalog # PA1016
  • Size 100 µg
Applications
Immunocytochemistry (1:50-1:200), Immunofluorescence (1:50-1:200), Immunoprecipitation (3 µg), Western Blot (1:500-1:2000)
Alternate names:
Stress-responsive protein 27; Heat shock 27 kDa protein; Estrogen-regulated 24 kDa protein; 28 kDa heat shock protein
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA1016 349.00
Check your price
  • Phospho-HSP27 (Ser15) Polyclonal Antibody for Western Blot, IF, ICC, IHC, Flow and IP
  • Host Rabbit
  • Target Species Human, Rat
  • Conjugate Unconjugated
  • Catalog # PA1018
  • Size 100 µg
Applications
Flow Cytometry (3-5 µg x 10^6 cells), Immunocytochemistry (1:50-1:200), Immunofluorescence (1:50-1:200), Immunohistochemistry (1:50-1:200), Immunoprecipitation (3 µg), Western Blot (1:100-1:1000)
Alternate names:
Stress-responsive protein 27; Heat shock 27 kDa protein; Estrogen-regulated 24 kDa protein; 28 kDa heat shock protein
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA1018 309.00
Check your price
  • Phospho-HSP27 (Ser78) Polyclonal Antibody for Western Blot, IF, ICC, IHC (P) and Flow
  • Host Rabbit
  • Target Species Human, Non-human primate
  • Conjugate Unconjugated
  • Catalog # PA517662
  • Size 100 µL
Applications
Flow Cytometry (1:100), Immunocytochemistry (1:100-1:500), Immunofluorescence (1:100-1:500), Immunohistochemistry (Paraffin) (1:100), Western Blot (1:1000)
Alternate names:
Stress-responsive protein 27; Heat shock 27 kDa protein; Estrogen-regulated 24 kDa protein; 28 kDa heat shock protein
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA517662 309.00
Check your price
  • Phospho-HSP27 (Ser82) Polyclonal Antibody for Western Blot, IHC (P) and Flow
  • Host Rabbit
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # 44534G
  • Size 10 blots
Applications
Flow Cytometry (3-5 µg/million cells), Immunohistochemistry (Paraffin) (1:20-1:200), Western Blot (1-2 µg/ml)
Alternate names:
Stress-responsive protein 27; Heat shock 27 kDa protein; Estrogen-regulated 24 kDa protein; 28 kDa heat shock protein
Catalog # Size Price (AUD) Your Price (AUD) Qty
44534G 304.00
Check your price
  • HSP27 Polyclonal Antibody for Western Blot
  • Host Rabbit
  • Target Species Human, Mouse, Rat
  • Conjugate Unconjugated
  • Catalog # AHO1132
  • Size 100 µg
Applications
Western Blot (Assay Dependent)
Alternate names:
Stress-responsive protein 27; Heat shock 27 kDa protein; Estrogen-regulated 24 kDa protein; 28 kDa heat shock protein; p25; Heat shock 25 kDa protein; Growth-related 25 kDa protein
Catalog # Size Price (AUD) Your Price (AUD) Qty
AHO1132 309.00
Check your price
  • HSP27 Polyclonal Antibody for Western Blot and IHC
  • Host Rabbit
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # PA514021
  • Size 400 µL
Applications
Immunohistochemistry (1:10-1:50), Western Blot (1:1000)
Alternate names:
Stress-responsive protein 27; Heat shock 27 kDa protein; Estrogen-regulated 24 kDa protein; 28 kDa heat shock protein
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA514021 349.00
Check your price
  • HSP27 Polyclonal Antibody for Western Blot and IHC
  • Host Rabbit
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # PA514022
  • Size 400 µL
Applications
Immunohistochemistry (1:10-1:50), Western Blot (1:1000)
Alternate names:
Stress-responsive protein 27; Heat shock 27 kDa protein; Estrogen-regulated 24 kDa protein; 28 kDa heat shock protein
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA514022 349.00
Check your price
  • Phospho-HSP27 (Ser78) Polyclonal Antibody for DB
  • Host Rabbit
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # PA512779
  • Size 400 µL
Applications
Dot blot (1:500)
Alternate names:
Stress-responsive protein 27; Heat shock 27 kDa protein; Estrogen-regulated 24 kDa protein; 28 kDa heat shock protein
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA512779 349.00
Check your price
  • Phospho-HSP27 (Ser82) Polyclonal Antibody for Western Blot
  • Host Rabbit
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # PA125983
  • Size 50 µL
Applications
Western Blot (1:1000)
Alternate names:
Stress-responsive protein 27; Heat shock 27 kDa protein; Estrogen-regulated 24 kDa protein; 28 kDa heat shock protein
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA125983 375.00
Check your price
  • Phospho-HSP25 (Ser86) Polyclonal Antibody for Western Blot, IF, ICC and Flow
  • Host Rabbit
  • Target Species Human, Mouse
  • Conjugate Unconjugated
  • Catalog # 44536G
  • Size 10 blots
Applications
Western Blot (Assay Dependent)
Alternate names:
Stress-responsive protein 27; Heat shock 27 kDa protein; Estrogen-regulated 24 kDa protein; 28 kDa heat shock protein; p25; Heat shock 25 kDa protein; Growth-related 25 kDa protein
Catalog # Size Price (AUD) Your Price (AUD) Qty
44536G 304.00
Check your price
  • SAFB Monoclonal Antibody for Western Blot, IF, ICC, IHC (P) and IP
  • Host Mouse
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # MA191526
  • Size 100 µg
Applications
Immunocytochemistry (Assay dependent), Immunofluorescence (Assay-dependent), Immunohistochemistry (Paraffin) (Assay dependent), Immunoprecipitation (Assay dependent), Western Blot (0.5 µg/ml)
Alternate names:
HSP27 estrogen response element-TATA box-binding protein
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA191526 425.00
Check your price