1 - 14 of 14 products displayed
  • HSC70 Polyclonal Antibody for Western Blot, IF, ICC, IHC (P) and Flow
  • Host Rabbit
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # PA524624
  • Size 400µL
Applications
Flow Cytometry (1:10-1:50), Immunocytochemistry (1:10-1:50), Immunofluorescence (1:10-1:50), Immunohistochemistry (1:10-1:50), Western Blot (1:1000)
Alternate names:
Lipopolysaccharide-associated protein 1; Heat shock 70 kDa protein 8
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA524624 349.00
Check your price
  • HSC70 Polyclonal Antibody for Western Blot
  • Host Goat
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # PA518537
  • Size 100ug
Applications
Western Blot (1-3 ug/ml)
Alternate names:
Lipopolysaccharide-associated protein 1; Heat shock 70 kDa protein 8
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA518537 349.00
Check your price
  • HSC70 Monoclonal Antibody for Western Blot, IF, ICC, IHC (F) and IP
  • Host Mouse
  • Target Species Cat, Human, Mouse, Non-human primate, Rat
  • Conjugate Unconjugated
  • Catalog # MA3014
  • Size 50µL
Applications
Immunocytochemistry (1:20-1:200), Immunofluorescence (1:20-1:200), Immunohistochemistry (Frozen) (1:20-1:200), Immunoprecipitation (2µl), Western Blot (1:1000-1:5000)
Alternate names:
Heat shock 70 kDa protein 8; Lipopolysaccharide-associated protein 1
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA3014 375.00
Check your price
  • HSC70 Monoclonal Antibody for Western Blot, IF, ICC, Flow, IP and ELISA
  • Host Rat
  • Target Species Many
  • Conjugate Unconjugated
  • Catalog # MA126078
  • Size 100µg
Applications
ELISA (Assay Dependent), Flow Cytometry (Assay Dependent), Immunocytochemistry (5 µg/ml), Immunofluorescence (5 ug/ml), Immunoprecipitation (5 µg/ml), Western Blot (1 µg/ml)
Alternate names:
Heat shock 70 kDa protein 8; Lipopolysaccharide-associated protein 1
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA126078 425.00
Check your price
  • HSC70 Monoclonal Antibody for Western Blot and IHC
  • Host Mouse
  • Target Species Many
  • Conjugate Unconjugated
  • Catalog # MA110891
  • Size 100µg
Applications
Immunohistochemistry (5 ug/ml), Western Blot (1 ug/ml)
Alternate names:
Heat shock 70 kDa protein 8; Lipopolysaccharide-associated protein 1
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA110891 349.00
Check your price
  • HSC70 Polyclonal Antibody for Western Blot, IF, ICC and IHC (P)
  • Host Rabbit
  • Target Species Human, Mouse
  • Conjugate Unconjugated
  • Catalog # PA527337
  • Size 100µL
Applications
Immunocytochemistry (1:100-1:1000), Immunofluorescence (1:100-1:1000), Immunohistochemistry (Paraffin) (1:100-1:1000), Western Blot (1:1000-1:10,000)
Alternate names:
Lipopolysaccharide-associated protein 1; Heat shock 70 kDa protein 8
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA527337 349.00
Check your price
  • HSC70 Polyclonal Antibody for Western Blot and IHC (P)
  • Host Rabbit
  • Target Species Human, Mouse, Pig
  • Conjugate Unconjugated
  • Catalog # PA529241
  • Size 100µL
Applications
Immunohistochemistry (Paraffin) (1:100-1:1000), Western Blot (1:1000-1:10,000)
Alternate names:
Lipopolysaccharide-associated protein 1; Heat shock 70 kDa protein 8
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA529241 349.00
Check your price
  • HSC70 Polyclonal Antibody for Western Blot, IF, ICC and IHC (P)
  • Host Rabbit
  • Target Species Human, Mouse
  • Conjugate Unconjugated
  • Catalog # PA529221
  • Size 100µL
Applications
Immunocytochemistry (1:100-1:1000), Immunofluorescence (1:100-1:1000), Immunohistochemistry (Paraffin) (1:100-1:1000), Western Blot (1:500-1:3000)
Alternate names:
Lipopolysaccharide-associated protein 1; Heat shock 70 kDa protein 8
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA529221 349.00
Check your price
  • HSC70 Monoclonal Antibody for Western Blot, IHC, Flow and IP
  • Host Mouse
  • Target Species Many
  • Conjugate Unconjugated
  • Catalog # MA110890
  • Size 100ug
Applications
Flow Cytometry (10 ug/ml), Immunohistochemistry (1-10 µg/ml), Immunoprecipitation (1-10 µg/ml), Western Blot (1 ug/ml)
Alternate names:
Heat shock 70 kDa protein 8; Lipopolysaccharide-associated protein 1
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA110890 375.00
Check your price
  • HSPH1 Polyclonal Antibody for Western Blot, IHC (P) and Flow
  • Host Rabbit
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # PA524619
  • Size 400µL
Applications
Flow Cytometry (1:10-1:50), Immunohistochemistry (1:50-1:100), Western Blot (1:1000)
Alternate names:
Heat shock 110 kDa protein; Antigen NY-CO-25
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA524619 349.00
Check your price
  • HSPH1 Polyclonal Antibody for Western Blot
  • Host Rabbit
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # PA512485
  • Size 400µL
Applications
Western Blot (1:1000)
Alternate names:
Heat shock 110 kDa protein; Antigen NY-CO-25
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA512485 349.00
Check your price
  • Apolipoprotein J Polyclonal Antibody for Western Blot and IHC (P)
  • Host Rabbit
  • Target Species Human, Mouse, Rat
  • Conjugate Unconjugated
  • Catalog # PA534651
  • Size 100µL
Applications
Immunohistochemistry (Paraffin) (1:100-1:1000), Western Blot (1:500-1:3000)
Alternate names:
Sulfated glycoprotein 2; Clustrin; Apolipoprotein J; Testosterone-repressed prostate message 2; NA1/NA2; Ku70-binding protein 1; Complement-associated protein SP-40,40; Complement cytolysis inhibitor; Aging-associated gene 4 protein; Testosterone repressed prostate message 2; Dimeric acid glycoprotein
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA534651 349.00
Check your price
  • Apolipoprotein J Monoclonal Antibody for Western Blot and IP
  • Host Rat
  • Target Species Mouse
  • Conjugate Unconjugated
  • Catalog # MA524052
  • Size 100µg
Applications
Immunoprecipitation (25 µg/ml), Western Blot (2 µg/ml)
Alternate names:
Sulfated glycoprotein 2; Clustrin; Apolipoprotein J
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA524052 275.00
Check your price
  • Apolipoprotein J Polyclonal Antibody for Western Blot, IHC (F), IP and ELISA
  • Host Goat
  • Target Species Mouse
  • Conjugate Unconjugated
  • Catalog # PA546931
  • Size 100µg
Applications
ELISA (0.1-0.4 µg/ml (Detection), 0.2-0.8 µg/ml (Capture)), Immunohistochemistry (Frozen) (5-15 µg/ml), Immunoprecipitation (25 µg/ml), Western Blot (0.1 µg/ml)
Alternate names:
Sulfated glycoprotein 2; Clustrin; Apolipoprotein J
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA546931 399.00
Check your price