1 - 2 of 2 products displayed
  • Tau Monoclonal Antibody for Western Blot, IF, ICC, IHC (P) and ELISA
  • Host Mouse
  • Target Species Bovine, Human
  • Conjugate Unconjugated
  • Catalog # MN1000
  • Size 100ug
Applications
ELISA (Assay Dependent), Immunocytochemistry (1:500-1:2000), Immunofluorescence (1:500-1:2000), Immunohistochemistry (Paraffin) (1:500-1:2000), Western Blot (Assay Dependent)
Alternate names:
Paired helical filament-tau; Neurofibrillary tangle protein
Catalog # Size Price (AUD) Your Price (AUD) Qty
MN1000 309.00
Check your price
  • Tau Monoclonal Antibody, Biotin conjugate for Western Blot, IHC and ELISA
  • Host Mouse
  • Target Species Bovine, Human
  • Conjugate Biotin
  • Catalog # MN1000B
  • Size 50ug
Applications
ELISA (Assay Dependent), Immunohistochemistry (Assay Dependent), Western Blot (Assay Dependent)
Alternate names:
Paired helical filament-tau; Neurofibrillary tangle protein
Catalog # Size Price (AUD) Your Price (AUD) Qty
MN1000B 309.00
Check your price