1 - 13 of 13 products displayed
  • HTRA2 Monoclonal Antibody for Western Blot, IHC and IHC (P)
  • Host Mouse
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # MA141040
  • Size 100µg
Applications
Immunohistochemistry (Paraffin) (1:10-1:500), Western Blot (2-3 µg/ml)
Alternate names:
Serine proteinase OMI; Serine protease 25; Omi stress-regulated endoprotease; High temperature requirement protein A2
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA141040 309.00
Check your price
  • HTRA2 Polyclonal Antibody for Western Blot and IHC (P)
  • Host Rabbit
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # PA529182
  • Size 100µL
Applications
Immunohistochemistry (Paraffin) (1:100-1:1000), Western Blot (1:500-1:3000)
Alternate names:
Serine proteinase OMI; Serine protease 25; Omi stress-regulated endoprotease; High temperature requirement protein A2
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA529182 349.00
Check your price
  • HTRA2 Polyclonal Antibody for Western Blot, IF, ICC and IHC
  • Host Rabbit
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # PA520064
  • Size 100ug
Applications
Immunocytochemistry (20 ug/ml), Immunofluorescence (20 ug/ml), Immunohistochemistry (10 µg/ml), Western Blot (0.5-1 ug/ml)
Alternate names:
Serine proteinase OMI; Serine protease 25; Omi stress-regulated endoprotease; High temperature requirement protein A2
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA520064 349.00
Check your price
  • HTRA2 Polyclonal Antibody for Western Blot, IF, ICC and IHC
  • Host Rabbit
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # PA520010
  • Size 100ug
Applications
Immunocytochemistry (20 ug/ml), Immunofluorescence (20 ug/ml), Immunohistochemistry (2 ug/ml), Western Blot (0.5-2 µg/ml)
Alternate names:
Serine proteinase OMI; Serine protease 25; Omi stress-regulated endoprotease; High temperature requirement protein A2
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA520010 349.00
Check your price
  • SFRS10 Polyclonal Antibody for Western Blot and IHC (P)
  • Host Rabbit
  • Target Species Human, Mouse, Rat
  • Conjugate Unconjugated
  • Catalog # PA530170
  • Size 100µL
Applications
Immunohistochemistry (Paraffin) (1:100-1:1000), Western Blot (1:500-1:3000)
Alternate names:
Transformer-2 protein homolog B; Splicing factor, arginine/serine-rich 10; RA301; Silica-induced gene 41 protein
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA530170 349.00
Check your price
  • SFRS10 Polyclonal Antibody for Western Blot and IHC (P)
  • Host Rabbit
  • Target Species Bovine, Dog, Horse, Guinea pig, Human, Mouse, Rabbit, Rat, Zebrafish
  • Conjugate Unconjugated
  • Catalog # PA541788
  • Size 100µl
Applications
Immunohistochemistry (Paraffin) (4-8 µg/ml), Western Blot (5 µg/ml)
Alternate names:
Transformer-2 protein homolog B; Splicing factor, arginine/serine-rich 10; RA301; Silica-induced gene 41 protein
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA541788 309.00
Check your price
  • SFRS10 Polyclonal Antibody for Western Blot and IHC (P)
  • Host Rabbit
  • Target Species Bovine, Dog, Horse, Human, Mouse, Pig, Rabbit, Rat
  • Conjugate Unconjugated
  • Catalog # PA541787
  • Size 100µl
Applications
Immunohistochemistry (Paraffin) (4-8 µg/ml), Western Blot (1.25 µg/ml)
Alternate names:
Transformer-2 protein homolog B; Splicing factor, arginine/serine-rich 10; RA301; Silica-induced gene 41 protein
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA541787 309.00
Check your price
  • HAX1 Monoclonal Antibody for Western Blot
  • Host Mouse
  • Target Species Human, Rat
  • Conjugate Unconjugated
  • Catalog # MA515770
  • Size 100ug
Applications
Western Blot (1-2 ug/ml)
Alternate names:
HS1-binding protein 1; HS1-associating protein X-1
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA515770 349.00
Check your price
  • HAX1 Monoclonal Antibody for Western Blot
  • Host Mouse
  • Target Species Human, Mouse, Rat
  • Conjugate Unconjugated
  • Catalog # MA515769
  • Size 100ug
Applications
Western Blot (1-2 ug/ml)
Alternate names:
HS1-binding protein 1; HS1-associating protein X-1
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA515769 349.00
Check your price
  • HAX1 Monoclonal Antibody for Western Blot
  • Host Mouse
  • Target Species Human, Rat
  • Conjugate Unconjugated
  • Catalog # MA515771
  • Size 100ug
Applications
Western Blot (1-2 ug/ml)
Alternate names:
HS1-binding protein 1; HS1-associating protein X-1
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA515771 349.00
Check your price
  • HAX1 Polyclonal Antibody for Western Blot
  • Host Rabbit
  • Target Species Human, Mouse, Rat
  • Conjugate Unconjugated
  • Catalog # PA520649
  • Size 100ug
Applications
Western Blot (1-2 ug/ml)
Alternate names:
HS1-binding protein 1; HS1-associating protein X-1
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA520649 349.00
Check your price
  • HAX1 Polyclonal Antibody for Western Blot
  • Host Rabbit
  • Target Species Human, Mouse, Rat
  • Conjugate Unconjugated
  • Catalog # PA520650
  • Size 100ug
Applications
Western Blot (1-2 ug/ml)
Alternate names:
HS1-binding protein 1; HS1-associating protein X-1
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA520650 349.00
Check your price
  • OMI / HtrA2 Synthetic Peptide for BLOCK and Ctrl
  • Host N/A
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # PEP0130
  • Size 50ug
Applications
Blocking Assay (Assay Dependent), Control (Assay Dependent)
Alternate names:
Catalog # Size Price (AUD) Your Price (AUD) Qty
PEP0130 85.75
Check your price