1 - 30 of 30 products displayed
  • alpha Synuclein Monoclonal Antibody for Western Blot, IHC (F), IHC (P), ELISA and DB
  • Host Mouse
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # MA190346
  • Size 100µL
Applications
Dot blot (1:100-1:10,000), ELISA (Assay dependent), Immunohistochemistry (Paraffin, Frozen) (1:100-1:1000), Western Blot (1:100-1:10,000)
Alternate names:
Non-A4 component of amyloid precursor; Non-A beta component of AD amyloid
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA190346 449.00
Check your price
  • alpha Synuclein Monoclonal Antibody for Western Blot, IF and IHC
  • Host Mouse
  • Target Species Human, Rat
  • Conjugate Unconjugated
  • Catalog # 391800
  • Size 100µg
Applications
Immunofluorescence (Assay Dependent), Immunohistochemistry (Assay Dependent), Western Blot (Assay Dependent)
Alternate names:
Non-A4 component of amyloid precursor; Non-A beta component of AD amyloid
Catalog # Size Price (AUD) Your Price (AUD) Qty
391800 309.00
Check your price
  • alpha Synuclein Monoclonal Antibody for Western Blot, IF, ICC, IHC (P), Flow and ELISA
  • Host Mouse
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # AHB0261
  • Size 100µg
Applications
ELISA (Assay Dependent), Flow Cytometry (3-5 µg/million cells), Immunocytochemistry (1 µg/ml), Immunofluorescence (1 µg/ml), Immunohistochemistry (Paraffin) (1:10-1:50), Western Blot (Assay Dependent)
Alternate names:
Non-A4 component of amyloid precursor; Non-A beta component of AD amyloid
Catalog # Size Price (AUD) Your Price (AUD) Qty
AHB0261 306.00
Check your price
  • alpha Synuclein Monoclonal Antibody for Western Blot, IF, IHC and ELISA
  • Host Mouse
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # 358300
  • Size 100µg
Applications
ELISA (Assay Dependent), Immunofluorescence (Assay Dependent), Immunohistochemistry (Assay Dependent), Western Blot (Assay Dependent)
Alternate names:
Non-A4 component of amyloid precursor; Non-A beta component of AD amyloid
Catalog # Size Price (AUD) Your Price (AUD) Qty
358300 304.00
Check your price
  • alpha Synuclein Monoclonal Antibody for Western Blot, IHC (P) and ELISA
  • Host Mouse
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # MA112874
  • Size 100µg
Applications
ELISA (Assay Dependent), Immunohistochemistry (Paraffin) (Assay Dependent), Western Blot (0.5-1.0 µg/ml)
Alternate names:
Non-A4 component of amyloid precursor; Non-A beta component of AD amyloid
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA112874 399.00
Check your price
  • alpha Synuclein Monoclonal Antibody for Western Blot, IHC and ELISA
  • Host Mouse
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # 328100
  • Size 100µg
Applications
ELISA (Assay Dependent), Immunohistochemistry (Assay Dependent), Western Blot (1-3 ug/ml)
Alternate names:
Non-A4 component of amyloid precursor; Non-A beta component of AD amyloid
Catalog # Size Price (AUD) Your Price (AUD) Qty
328100 310.00
Check your price
  • alpha Thrombin for Ctrl
  • Host Human
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # RP43100
  • Size 100µg
Applications
Control (Assay Dependent)
Alternate names:
Coagulation factor II
Catalog # Size Price (AUD) Your Price (AUD) Qty
RP43100 122.00
Check your price
  • alpha Synuclein Polyclonal Antibody for IHC (P)
  • Host Rabbit
  • Target Species Human, Rat
  • Conjugate Unconjugated
  • Catalog # PA121121
  • Size 500µL
Applications
Immunohistochemistry (Paraffin) (1:100)
Alternate names:
Non-A4 component of amyloid precursor; Non-A beta component of AD amyloid
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA121121 375.00
Check your price
  • alpha Synuclein Polyclonal Antibody for Western Blot, IF, ICC, IHC (P) and Flow
  • Host Rabbit
  • Target Species Human, Rat
  • Conjugate Unconjugated
  • Catalog # PA516738
  • Size 500µL
Applications
Immunohistochemistry (Paraffin) (1:800)
Alternate names:
Non-A4 component of amyloid precursor; Non-A beta component of AD amyloid
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA516738 309.00
Check your price
  • alpha-1 Antitrypsin Polyclonal Antibody for Western Blot
  • Host Rabbit
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # PA527311
  • Size 100µL
Applications
Western Blot (1:500-1:3000)
Alternate names:
Serpin A1; Alpha-1-antiproteinase; Alpha-1 protease inhibitor
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA527311 349.00
Check your price
  • alpha Synuclein Monoclonal Antibody for Western Blot, IHC (F), IHC (P) and ELISA
  • Host Mouse
  • Target Species Human, Mouse, Rat
  • Conjugate Unconjugated
  • Catalog # MA190342
  • Size 100µL
Applications
ELISA (Assay Dependent), Immunohistochemistry (Paraffin, Frozen) (1:100-1:1000), Western Blot (1:100-1:10,000)
Alternate names:
Non-A4 component of amyloid precursor; Non-A beta component of AD amyloid
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA190342 425.00
Check your price
  • Synuclein alpha/beta Monoclonal Antibody for Western Blot, IF, ICC, IHC, Flow, IP and ELISA
  • Host Mouse
  • Target Species Human, Mouse
  • Conjugate Unconjugated
  • Catalog # 328200
  • Size 100µg
Applications
ELISA (Assay Dependent), Flow Cytometry (3-5 ug x 10^6 cells), Immunocytochemistry (1-2 ug/ml), Immunofluorescence (1-2 ug/ml), Immunohistochemistry (Assay Dependent), Immunoprecipitation (Assay Dependent), Western Blot (1-3 ug/ml)
Alternate names:
Non-A4 component of amyloid precursor; Non-A beta component of AD amyloid
Catalog # Size Price (AUD) Your Price (AUD) Qty
328200 305.00
Check your price
  • Phospho-alpha Synuclein (Tyr136) Polyclonal Antibody for Western Blot, IF and ICC
  • Host Rabbit
  • Target Species Human, Mouse, Rat
  • Conjugate Unconjugated
  • Catalog # PA537743
  • Size 100µl
Applications
Immunocytochemistry (1:100-1:200), Immunofluorescence (1:100-1:200), Western Blot (1:500-1:1000)
Alternate names:
Non-A4 component of amyloid precursor; Non-A beta component of AD amyloid
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA537743 309.00
Check your price
  • Phospho-alpha Synuclein (Tyr125) Polyclonal Antibody for Western Blot, IF and ICC
  • Host Rabbit
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # PA537741
  • Size 100µl
Applications
Immunocytochemistry (1:100-1:200), Immunofluorescence (1:100-1:200), Western Blot (1:500-1:1000)
Alternate names:
Non-A4 component of amyloid precursor; Non-A beta component of AD amyloid
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA537741 309.00
Check your price
  • Phospho-alpha Synuclein (Tyr133) Polyclonal Antibody for Western Blot, IF and ICC
  • Host Rabbit
  • Target Species Human, Mouse, Rat
  • Conjugate Unconjugated
  • Catalog # PA537742
  • Size 100µl
Applications
Immunocytochemistry (1:100-1:200), Immunofluorescence (1:100-1:200), Western Blot (1:500-1:1000)
Alternate names:
Non-A4 component of amyloid precursor; Non-A beta component of AD amyloid
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA537742 309.00
Check your price
  • Phospho-alpha Synuclein (Ser129) Polyclonal Antibody for Western Blot
  • Host Rabbit
  • Target Species Human, Mouse, Rat
  • Conjugate Unconjugated
  • Catalog # PA537740
  • Size 100µl
Applications
Western Blot (1:500-1:1000)
Alternate names:
Non-A4 component of amyloid precursor; Non-A beta component of AD amyloid
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA537740 309.00
Check your price
  • alpha Synuclein Polyclonal Antibody for Western Blot and IHC
  • Host Rabbit
  • Target Species Mouse, Rat, Human
  • Conjugate Unconjugated
  • Catalog # PA118264
  • Size 100µL
Applications
Immunohistochemistry (1:500-1:3000), Western Blot (1:500-1:3000)
Alternate names:
Non-A4 component of amyloid precursor; Non-A beta component of AD amyloid
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA118264 399.00
Check your price
  • alpha Synuclein Polyclonal Antibody for IHC (P)
  • Host Sheep
  • Target Species Human, Rat
  • Conjugate Unconjugated
  • Catalog # PA118258
  • Size 50µg
Applications
Immunohistochemistry (Paraffin) (1 µg/ml)
Alternate names:
Non-A4 component of amyloid precursor; Non-A beta component of AD amyloid
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA118258 375.00
Check your price
  • alpha Synuclein Polyclonal Antibody for Western Blot and IHC
  • Host Rabbit
  • Target Species Human, Rat
  • Conjugate Unconjugated
  • Catalog # PA118253
  • Size 100µL
Applications
Immunohistochemistry (1:1,000), Western Blot (1:1,000)
Alternate names:
Non-A4 component of amyloid precursor; Non-A beta component of AD amyloid
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA118253 349.00
Check your price
  • alpha-1 Antitrypsin Monoclonal Antibody for IHC (F) and IHC (P)
  • Host Mouse
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # MA190438
  • Size 500µL
Applications
Immunohistochemistry (Frozen) (Neat), Immunohistochemistry (Paraffin) (Neat)
Alternate names:
Serpin A1; Alpha-1-antiproteinase; Alpha-1 protease inhibitor
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA190438 349.00
Check your price
  • alpha-1 Antitrypsin Polyclonal Antibody for Western Blot and ELISA
  • Host Chicken
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # PA19529
  • Size 500ug
Applications
ELISA (Assay Dependent), Western Blot (1:1000)
Alternate names:
Serpin A1; Alpha-1-antiproteinase; Alpha-1 protease inhibitor
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA19529 199.00
Check your price
  • alpha Synuclein Polyclonal Antibody for Western Blot, IHC (F) and IHC (P)
  • Host Sheep
  • Target Species Human, Rat
  • Conjugate Unconjugated
  • Catalog # PA118256
  • Size 100µg
Applications
Immunohistochemistry (Paraffin, Frozen) (3 µg/ml), Western Blot (1 µg/ml)
Alternate names:
Non-A4 component of amyloid precursor; Non-A beta component of AD amyloid
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA118256 375.00
Check your price
  • alpha Synuclein Oligoclonal Antibody for Western Blot, IF and ICC
  • Host Rabbit
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # 710110
  • Size 100µg
Applications
Immunocytochemistry (1:500-1:5000), Immunofluorescence (1:500-1:5000), Western Blot (1:500-1:5000)
Alternate names:
Non-A4 component of amyloid precursor; Non-A beta component of AD amyloid
Catalog # Size Price (AUD) Your Price (AUD) Qty
710110 309.00
Check your price
  • alpha-1 Antitrypsin Polyclonal Antibody, HRP conjugate for ELISA
  • Host Sheep
  • Target Species Human
  • Conjugate HRP
  • Catalog # PA184595
  • Size 1mL
Applications
ELISA (1:6400-1:12,800)
Alternate names:
Serpin A1; Alpha-1-antiproteinase; Alpha-1 protease inhibitor
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA184595 389.00
Check your price
  • alpha-1 Antitrypsin Monoclonal Antibody for Flow and ELISA
  • Host Mouse
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # MIA1102
  • Size 1mg
Applications
ELISA (Assay Dependent), Flow Cytometry (Assay Dependent)
Alternate names:
Serpin A1; Alpha-1-antiproteinase; Alpha-1 protease inhibitor
Catalog # Size Price (AUD) Your Price (AUD) Qty
MIA1102 309.00
Check your price
  • alpha Synuclein Monoclonal Antibody for Western Blot, IF, ICC, IHC (P) and IP
  • Host Mouse
  • Target Species Human, Rat
  • Conjugate Unconjugated
  • Catalog # MA512272
  • Size 500µL
Applications
Immunocytochemistry (1:20-1:200), Immunofluorescence (1:20-1:200), Immunohistochemistry (Paraffin) (1:200-1:400), Immunoprecipitation (Assay Dependent), Western Blot (Assay Dependent)
Alternate names:
Non-A4 component of amyloid precursor; Non-A beta component of AD amyloid
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA512272 309.00
Check your price
  • alpha Synuclein Monoclonal Antibody for Western Blot, IF, ICC, IHC (P) and Flow
  • Host Rabbit
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # 701085
  • Size 100µg
Applications
Flow Cytometry (1:100-1:500), Immunocytochemistry (1:500-1:5000), Immunofluorescence (1:500-1:5000), Immunohistochemistry (Paraffin) (1:100-1:1000), Western Blot (1:500-1:5000)
Alternate names:
Non-A4 component of amyloid precursor; Non-A beta component of AD amyloid
Catalog # Size Price (AUD) Your Price (AUD) Qty
701085 304.00
Check your price
  • alpha Synuclein Polyclonal Antibody for Western Blot
  • Host Rabbit
  • Target Species Human, Mouse
  • Conjugate Unconjugated
  • Catalog # PA513401
  • Size 400µL
Applications
Western Blot (1:1000)
Alternate names:
Non-A4 component of amyloid precursor; Non-A beta component of AD amyloid
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA513401 349.00
Check your price
  • alpha-1 Antitrypsin Monoclonal Antibody for Western Blot
  • Host Mouse
  • Target Species Human, Mouse
  • Conjugate Unconjugated
  • Catalog # MA515521
  • Size 100µL
Applications
Western Blot (1/500-1/2000)
Alternate names:
Serpin A1; Alpha-1-antiproteinase; Alpha-1 protease inhibitor; Serine protease inhibitor A1a; Serine protease inhibitor 1-1; Alpha-1 protease inhibitor 1
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA515521 349.00
Check your price
  • alpha Synuclein Polyclonal Antibody for Western Blot and IHC
  • Host Rabbit
  • Target Species Human, Mouse
  • Conjugate Unconjugated
  • Catalog # PA513397
  • Size 400µL
Applications
Immunohistochemistry (1:50-1:100), Western Blot (1:1000)
Alternate names:
Non-A4 component of amyloid precursor; Non-A beta component of AD amyloid
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA513397 349.00
Check your price