1 - 30 of 30 products displayed
  • FPR3 Polyclonal Antibody for ICC and IHC (P)
  • Host Rabbit
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # PA533541
  • Size 50 ug
Applications
Immunocytochemistry (Assay Dependent), Immunohistochemistry (Paraffin) (3 µg/ml)
Alternate names:
Formyl peptide receptor-like 2; FMLP-related receptor II
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA533541 349.00
Check your price
  • FPR3 Polyclonal Antibody for IHC (P)
  • Host Rabbit
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # PA533540
  • Size 50 ug
Applications
Immunohistochemistry (Paraffin) (8 µg/ml)
Alternate names:
Formyl peptide receptor-like 2; FMLP-related receptor II
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA533540 349.00
Check your price
  • REG3A Polyclonal Antibody for Western Blot
  • Host Rabbit
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # PA528780
  • Size 100 µL
Applications
Western Blot (1:500-1:3000)
Alternate names:
Regenerating islet-derived protein III-alpha; Pancreatitis-associated protein 1; Human proislet peptide; Hepatointestinal pancreatic protein
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA528780 349.00
Check your price
  • REG3A Polyclonal Antibody for Western Blot and IHC (P)
  • Host Rabbit
  • Target Species Human, Non-human primate
  • Conjugate Unconjugated
  • Catalog # PA523341
  • Size 100 ug
Applications
Immunohistochemistry (Paraffin) (2.5-5 µg/ml), Western Blot (1-3 µg/ml)
Alternate names:
Regenerating islet-derived protein III-alpha; Pancreatitis-associated protein 1; Human proislet peptide; Hepatointestinal pancreatic protein
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA523341 349.00
Check your price
  • REG3A Polyclonal Antibody for Western Blot and Flow
  • Host Rabbit
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # PA526219
  • Size 400 µL
Applications
Flow Cytometry (1:10-1:50), Western Blot (1:1000)
Alternate names:
Regenerating islet-derived protein III-alpha; Pancreatitis-associated protein 1; Human proislet peptide; Hepatointestinal pancreatic protein
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA526219 349.00
Check your price
  • THOP1 Polyclonal Antibody for Western Blot
  • Host Rabbit
  • Target Species Human, Mouse
  • Conjugate Unconjugated
  • Catalog # PA530229
  • Size 100 µL
Applications
Western Blot (1:500-1:3000)
Alternate names:
MP78; Endopeptidase 24.15
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA530229 349.00
Check your price
  • CDH17 Polyclonal Antibody for Western Blot, IF and IHC (P)
  • Host Rabbit
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # PA536295
  • Size 100 µl
Applications
Immunofluorescence (1:50-1:200), Immunohistochemistry (Paraffin) (1:50-1:200), Western Blot (1:500-1:1000)
Alternate names:
Liver-intestine cadherin; Intestinal peptide-associated transporter HPT-1
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA536295 309.00
Check your price
  • THOP1 Monoclonal Antibody for Western Blot and IP
  • Host Mouse
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # MA524174
  • Size 100 µg
Applications
Immunoprecipitation (25 µg/ml), Western Blot (1 µg/ml)
Alternate names:
MP78; Endopeptidase 24.15
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA524174 275.00
Check your price
  • THOP1 Polyclonal Antibody for Western Blot, IHC and IP
  • Host Sheep
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # PA548031
  • Size 100 µg
Applications
Immunohistochemistry (5-15 µg/ml), Immunoprecipitation (25 µg/ml), Western Blot (0.1 µg/ml)
Alternate names:
MP78; Endopeptidase 24.15
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA548031 309.00
Check your price
  • FPR3 Polyclonal Antibody for IHC (P)
  • Host Rabbit
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # PA132544
  • Size 25 µg
Applications
Immunohistochemistry (Paraffin) (1:100-1:200)
Alternate names:
Formyl peptide receptor-like 2; FMLP-related receptor II
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA132544 399.00
Check your price
  • Aquaporin 10 Polyclonal Antibody for Western Blot
  • Host Rabbit
  • Target Species Bovine, Human, Rabbit, Rat
  • Conjugate Unconjugated
  • Catalog # PA543530
  • Size 100 µl
Applications
Western Blot (0.2-1 µg/ml)
Alternate names:
Small intestine aquaporin; Aquaglyceroporin-10
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA543530 309.00
Check your price
  • PYY Polyclonal Antibody for IHC (F) and IHC (P)
  • Host Rabbit
  • Target Species Human, Rat
  • Conjugate Unconjugated
  • Catalog # PA136142
  • Size 50 µL
Applications
Immunohistochemistry (Frozen) (1:1000-1:5000), Immunohistochemistry (Paraffin) (1:1000-1:5000)
Alternate names:
Peptide tyrosine tyrosine; PYY-I
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA136142 900.00
Check your price
  • PYY Polyclonal Antibody, Biotin conjugate for ELISA
  • Host Chicken
  • Target Species Human, Mouse, Rat
  • Conjugate Biotin
  • Catalog # PA19590
  • Size 100 µL
Applications
ELISA (1:2000-1:10000)
Alternate names:
Peptide tyrosine tyrosine; PYY-I
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA19590 375.00
Check your price
  • PYY Polyclonal Antibody, Biotin conjugate for ELISA
  • Host Chicken
  • Target Species Human, Mouse, Rat
  • Conjugate Biotin
  • Catalog # PA19589
  • Size 100 µL
Applications
ELISA (1:2000-1:10000)
Alternate names:
Peptide tyrosine tyrosine; PYY-I
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA19589 375.00
Check your price
  • PYY Polyclonal Antibody for ELISA
  • Host Chicken
  • Target Species Human, Mouse, Rat
  • Conjugate Unconjugated
  • Catalog # PA19582
  • Size 100 µL
Applications
ELISA (1:2000-1:5000)
Alternate names:
Peptide tyrosine tyrosine; PYY-I
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA19582 375.00
Check your price
  • PYY Polyclonal Antibody for ELISA
  • Host Chicken
  • Target Species Human, Mouse, Rat
  • Conjugate Unconjugated
  • Catalog # PA19581
  • Size 100 µL
Applications
ELISA (1:2000-1:5000)
Alternate names:
Peptide tyrosine tyrosine; PYY-I
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA19581 375.00
Check your price
  • CDH17 Polyclonal Antibody for Western Blot
  • Host Chicken
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # PA127949
  • Size 50µg
Applications
Western Blot (1:2000)
Alternate names:
Liver-intestine cadherin; Intestinal peptide-associated transporter HPT-1
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA127949 425.00
Check your price
  • PYY Polyclonal Antibody for IHC (F) and IHC (P)
  • Host Guinea Pig
  • Target Species Cat, Mouse, Rat
  • Conjugate Unconjugated
  • Catalog # PA136027
  • Size 50 µL
Applications
Immunohistochemistry (Paraffin, Frozen) (1:1,500-1:2,500)
Alternate names:
Peptide tyrosine tyrosine
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA136027 550.00
Check your price
  • NPR3 Polyclonal Antibody for Western Blot, IF, ICC and IHC (P)
  • Host Rabbit
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # PA515394
  • Size 400 µL
Applications
Immunocytochemistry (1:200), Immunofluorescence (1:200), Immunohistochemistry (1:10-1:50), Western Blot (1:1000)
Alternate names:
Atrial natriuretic peptide receptor type C; Atrial natriuretic peptide clearance receptor
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA515394 349.00
Check your price
  • Amylin Polyclonal Antibody for IHC (P)
  • Host Rabbit
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # PA532261
  • Size 500 µL
Applications
Immunohistochemistry (Paraffin) (1:200)
Alternate names:
Insulinoma amyloid peptide; Diabetes-associated peptide; Amylin
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA532261 275.00
Check your price
  • Amylin Polyclonal Antibody for IHC (P)
  • Host Rabbit
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # PA137075
  • Size 1 mL
Applications
Immunohistochemistry (Paraffin) (1:200)
Alternate names:
Insulinoma amyloid peptide; Diabetes-associated peptide; Amylin
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA137075 399.00
Check your price
  • Ghrelin Polyclonal Antibody for ELISA
  • Host Chicken
  • Target Species Human, Mouse
  • Conjugate Unconjugated
  • Catalog # PA19595
  • Size 200 µg
Applications
ELISA (1:500-1:4000)
Alternate names:
Protein M46; Motilin-related peptide; Growth hormone-releasing peptide; Growth hormone secretagogue
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA19595 309.00
Check your price
  • HLA-ABC Monoclonal Antibody, PE conjugate for Flow
  • Host Mouse
  • Target Species Bovine, Cat, Human, Non-human primate
  • Conjugate PE
  • Catalog # MA119662
  • Size 100 µg
Applications
Flow Cytometry (1:50)
Alternate names:
MHC class I antigen A*1; MHC class I antigen B*8; MHC class I antigen A*3; MHC class I antigen B*7; MHC class I antigen Cw*7
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA119662 375.00
Check your price
  • HLA-ABC Monoclonal Antibody for Flow, IP and FN
  • Host Mouse
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # MA170111
  • Size 500 ug
Applications
Flow Cytometry (1 ug/1 million cells), Functional Assay (Assay Dependent), Immunoprecipitation (Assay Dependent)
Alternate names:
MHC class I antigen A*1; MHC class I antigen B*8; MHC class I antigen A*3; MHC class I antigen B*7; MHC class I antigen Cw*7
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA170111 309.00
Check your price
  • HLA-ABC Monoclonal Antibody for Western Blot, ICC, IHC (F), Flow, IP, ELISA and FN
  • Host Mouse
  • Target Species Bovine, Cat, Human, Non-human primate
  • Conjugate Unconjugated
  • Catalog # MA119027
  • Size 100 µg
Applications
ELISA (Assay Dependent), Flow Cytometry (Assay Dependent), Functional Assay (Assay Dependent), Immunocytochemistry (Assay Dependent), Immunohistochemistry (Frozen) (Assay Dependent), Immunoprecipitation (Assay Dependent), Western Blot (Assay Dependent)
Alternate names:
MHC class I antigen A*1; MHC class I antigen B*8; MHC class I antigen A*3; MHC class I antigen B*7; MHC class I antigen Cw*7
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA119027 349.00
Check your price
  • HLA-ABC Monoclonal Antibody for Western Blot, IF, ICC, IHC (F) and Flow
  • Host Mouse
  • Target Species Human, Mouse
  • Conjugate Unconjugated
  • Catalog # MA511723
  • Size 500 µL
Applications
Flow Cytometry (0.5 ug/test), Immunocytochemistry (1:10-1:200), Immunofluorescence (1:10-1:200), Immunohistochemistry (Frozen) (Assay Dependent), Western Blot (1:10-1:200)
Alternate names:
MHC class I antigen A*1; MHC class I antigen B*8; MHC class I antigen A*3; MHC class I antigen B*7; MHC class I antigen Cw*7
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA511723 299.00
Check your price
  • CGRP Monoclonal Antibody for IHC (P) and ELISA
  • Host Mouse
  • Target Species Human, Rat
  • Conjugate Unconjugated
  • Catalog # ABS0260502
  • Size 200 µL
Applications
ELISA (1:2000), Immunohistochemistry (Paraffin) (1:400)
Alternate names:
Calcitonin gene-related peptide I; Alpha-type CGRP
Catalog # Size Price (AUD) Your Price (AUD) Qty
ABS0260502 550.00
Check your price
  • Calcitonin Monoclonal Antibody for IHC (P)
  • Host Rabbit
  • Target Species Dog, Horse, Human, Mouse, Non-human primate, Sheep, Pig, Rat
  • Conjugate Unconjugated
  • Catalog # MA514552
  • Size 500 µL
Applications
Immunohistochemistry (Paraffin) (1:200)
Alternate names:
Calcitonin gene-related peptide I; Alpha-type CGRP
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA514552 349.00
Check your price
  • Calcitonin Monoclonal Antibody for IHC (P)
  • Host Rabbit
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # MA516325
  • Size 500 µL
Applications
Immunohistochemistry (Paraffin) (1:100)
Alternate names:
Calcitonin gene-related peptide I; Alpha-type CGRP
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA516325 375.00
Check your price
  • GRPR Polyclonal Antibody for IHC (P)
  • Host Rabbit
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # OPA115619
  • Size 25 µg
Applications
Immunohistochemistry (Paraffin) (Assay dependent)
Alternate names:
GRP-preferring bombesin receptor
Catalog # Size Price (AUD) Your Price (AUD) Qty
OPA115619 499.00
Check your price