1 - 2 of 2 products displayed
  • CD133 Monoclonal Antibody for Western Blot
  • Host Mouse
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # MA518323
  • Size 100µl
Applications
Western Blot (1:500-1:1000)
Alternate names:
Prominin-like protein 1; Antigen AC133
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA518323 399.00
Check your price
  • CD133 Polyclonal Antibody for Western Blot, IF and ICC
  • Host Rabbit
  • Target Species Human, Mouse, Rat
  • Conjugate Unconjugated
  • Catalog # PA538014
  • Size 100µg
Applications
Immunocytochemistry (2.5 µg/ml), Immunofluorescence (20 µg/ml), Western Blot (5 µg/ml)
Alternate names:
Prominin-like protein 1; Antigen AC133; Antigen AC133 homolog
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA538014 309.00
Check your price