1 - 30 of 30 products displayed
  • CDK4 Polyclonal Antibody for Western Blot, IF, ICC and IHC (P)
  • Host Rabbit
  • Target Species Human, Mouse
  • Conjugate Unconjugated
  • Catalog # PA527827
  • Size 100µL
Applications
Immunocytochemistry (1:100-1:1000), Immunofluorescence (1:100-1:1000), Immunohistochemistry (Paraffin) (1:100-1:1000), Western Blot (1:500-1:3000)
Alternate names:
PSK-J3; Cell division protein kinase 4; CRK3
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA527827 349.00
Check your price
  • CDK4 Monoclonal Antibody for Western Blot, IF, Flow and IP
  • Host Mouse
  • Target Species Human, Mouse, Rat
  • Conjugate Unconjugated
  • Catalog # MA513720
  • Size 500µL
Applications
Flow Cytometry (3-5 µg x 10^6 cells), Immunofluorescence (Assay Dependent), Immunoprecipitation (2 µg/ml), Western Blot (1-2 µg/ml)
Alternate names:
PSK-J3; Cell division protein kinase 4; CRK3
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA513720 349.00
Check your price
  • CDK4 Monoclonal Antibody for Western Blot, IF, Flow and IP
  • Host Mouse
  • Target Species Human, Mouse, Pig, Rat
  • Conjugate Unconjugated
  • Catalog # MA512984
  • Size 500µL
Applications
Flow Cytometry (3-5 µg x 10^6 cells), Immunofluorescence (Assay Dependent), Immunoprecipitation (2 µg/ml), Western Blot (1-2 µg/ml)
Alternate names:
PSK-J3; Cell division protein kinase 4; CRK3
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA512984 349.00
Check your price
  • CDK4 Monoclonal Antibody, Biotin conjugate for Western Blot, IF and IP
  • Host Mouse
  • Target Species Mouse, Pig, Rat, Human
  • Conjugate Biotin
  • Catalog # MA512981
  • Size 500µL
Applications
Immunofluorescence (Assay Dependent), Immunoprecipitation (2 µg/ml), Western Blot (1-2 µg/ml)
Alternate names:
PSK-J3; Cell division protein kinase 4; CRK3
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA512981 399.00
Check your price
  • CDK4 Monoclonal Antibody for Western Blot, IF, Flow and IP
  • Host Mouse
  • Target Species Human, Mouse, Rat
  • Conjugate Unconjugated
  • Catalog # AHZ0202
  • Size 500µl
Applications
Flow Cytometry (Assay-Dependent), Immunofluorescence (Assay Dependent), Immunoprecipitation (Assay-Dependent), Western Blot (Assay-Dependent)
Alternate names:
PSK-J3; Cell division protein kinase 4; CRK3
Catalog # Size Price (AUD) Your Price (AUD) Qty
AHZ0202 312.00
Check your price
  • CDK4 Monoclonal Antibody for Western Blot, IF and IP
  • Host Mouse
  • Target Species Human, Mouse, Rat
  • Conjugate Unconjugated
  • Catalog # MA112299
  • Size 100µg
Applications
Immunofluorescence (Assay Dependent), Immunoprecipitation (Assay Dependent), Western Blot (Assay Dependent)
Alternate names:
PSK-J3; Cell division protein kinase 4; CRK3
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA112299 309.00
Check your price
  • CDK4 Polyclonal Antibody for Western Blot, IF, ICC, IHC (P) and Flow
  • Host Rabbit
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # PA514445
  • Size 400µL
Applications
Flow Cytometry (1:10-1:50), Immunocytochemistry (1:10-1:50), Immunofluorescence (1:10-1:50), Immunohistochemistry (1:50-1:100), Western Blot (1:1000)
Alternate names:
PSK-J3; Cell division protein kinase 4
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA514445 349.00
Check your price
  • CDK4 Polyclonal Antibody for ELISA
  • Host Sheep
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # PA175092
  • Size 500µg
Applications
ELISA (Assay Dependent)
Alternate names:
PSK-J3; Cell division protein kinase 4
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA175092 1,400.00
Check your price
  • CDK4 Polyclonal Antibody for Western Blot and IP
  • Host Rabbit
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # PA112580
  • Size 100µg
Applications
Immunoprecipitation (Assay Dependent), Western Blot (Assay Dependent)
Alternate names:
PSK-J3; Cell division protein kinase 4
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA112580 309.00
Check your price
  • CDK4 Monoclonal Antibody for Western Blot, IHC (F) and IP
  • Host Mouse
  • Target Species Human, Mouse, Rat
  • Conjugate Unconjugated
  • Catalog # MA182085
  • Size 100µg
Applications
Immunohistochemistry (Frozen) (Assay Dependent), Immunoprecipitation (Assay Dependent), Western Blot (Assay Dependent)
Alternate names:
PSK-J3; Cell division protein kinase 4; CRK3
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA182085 550.00
Check your price
  • CDK4 Oligoclonal Antibody for Western Blot
  • Host Rabbit
  • Target Species Human, Rat
  • Conjugate Unconjugated
  • Catalog # 710017
  • Size 100µg
Applications
Western Blot (1-5 ug/ml)
Alternate names:
PSK-J3; Cell division protein kinase 4
Catalog # Size Price (AUD) Your Price (AUD) Qty
710017 309.00
Check your price
  • CDKN2A Polyclonal Antibody for Western Blot, IF, ICC and IHC
  • Host Rabbit
  • Target Species Human, Mouse, Rat
  • Conjugate Unconjugated
  • Catalog # PA520379
  • Size 100ug
Applications
Immunocytochemistry (20 ug/ml), Immunofluorescence (20 ug/ml), Immunohistochemistry (10 µg/ml), Western Blot (1-2 ug/ml)
Alternate names:
p16-INK4a; Cyclin-dependent kinase 4 inhibitor A; p14ARF; Cyclin-dependent kinase inhibitor 2A; Alternative reading frame; p19ARF; Multiple tumor suppressor 1
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA520379 349.00
Check your price
  • p19 INK4d Monoclonal Antibody for Western Blot, IP and ELISA
  • Host Mouse
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # 378700
  • Size 100µg
Applications
ELISA (Assay Dependent), Immunoprecipitation (Assay Dependent), Western Blot (Assay Dependent)
Alternate names:
p19-INK4d
Catalog # Size Price (AUD) Your Price (AUD) Qty
378700 304.00
Check your price
  • p19 INK4d Polyclonal Antibody for Western Blot, IHC (P) and Flow
  • Host Rabbit
  • Target Species Human, Mouse
  • Conjugate Unconjugated
  • Catalog # PA526413
  • Size 400µL
Applications
Flow Cytometry (1:10-1:50), Immunohistochemistry (1:10-1:50), Western Blot (1:1000)
Alternate names:
p19-INK4d
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA526413 349.00
Check your price
  • SERTAD1 Polyclonal Antibody for Western Blot and IHC (P)
  • Host Rabbit
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # PA528460
  • Size 100µL
Applications
Immunohistochemistry (Paraffin) (1:100-1:1000), Western Blot (1:500-1:3000)
Alternate names:
Transcriptional regulator interacting with the PHD-bromodomain 1; CDK4-binding protein p34SEI1
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA528460 349.00
Check your price
  • CDKN2A Polyclonal Antibody for IHC (P)
  • Host Rabbit
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # PA128218
  • Size 7mL
Applications
Immunohistochemistry (Paraffin) (Assay dependent)
Alternate names:
p14ARF; Cyclin-dependent kinase inhibitor 2A; Alternative reading frame; p16-INK4a; Multiple tumor suppressor 1; Cyclin-dependent kinase 4 inhibitor A
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA128218 375.00
Check your price
  • CDKN2A Polyclonal Antibody for Western Blot, IF, ICC, IHC (F) and IHC (P)
  • Host Rabbit
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # PA129914
  • Size 500µL
Applications
Immunocytochemistry (Assay-dependent), Immunofluorescence (Assay dependent), Immunohistochemistry (Frozen) (1:50-1:100), Immunohistochemistry (Paraffin) (1:50-1:100), Western Blot (Assay dependent)
Alternate names:
p14ARF; Cyclin-dependent kinase inhibitor 2A; Alternative reading frame; p16-INK4a; Multiple tumor suppressor 1; Cyclin-dependent kinase 4 inhibitor A
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA129914 425.00
Check your price
  • SERTAD1 Polyclonal Antibody for Western Blot
  • Host Chicken
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # PA127780
  • Size 50µg
Applications
Western Blot (1:2000)
Alternate names:
Transcriptional regulator interacting with the PHD-bromodomain 1; CDK4-binding protein p34SEI1
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA127780 425.00
Check your price
  • CDKN2A Polyclonal Antibody for Western Blot, IF, ICC, IHC (F), IHC (P), Flow, IP and ELISA
  • Host Rabbit
  • Target Species Human, Mouse
  • Conjugate Unconjugated
  • Catalog # PA131409
  • Size 100µL
Applications
ELISA (1:100-1:2000), Flow Cytometry (1:400), Immunocytochemistry (1:50-1:200), Immunofluorescence (1:50-1:200), Immunohistochemistry (Frozen) (1:10-1:200), Immunohistochemistry (Paraffin) (1:10-1:200), Immunoprecipitation (Assay-dependent), Western Blot (1:100-1:2000)
Alternate names:
p16-INK4a; Cyclin-dependent kinase 4 inhibitor A; p14ARF; Cyclin-dependent kinase inhibitor 2A; Alternative reading frame; p19ARF; Multiple tumor suppressor 1
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA131409 425.00
Check your price
  • CDKN2A Polyclonal Antibody for Western Blot
  • Host Rabbit
  • Target Species Human, Mouse, Rat
  • Conjugate Unconjugated
  • Catalog # PA131212
  • Size 100µg
Applications
Western Blot (1:500)
Alternate names:
p16-INK4a; Cyclin-dependent kinase 4 inhibitor A; p14ARF; Cyclin-dependent kinase inhibitor 2A; Alternative reading frame; p19ARF; Multiple tumor suppressor 1
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA131212 449.00
Check your price
  • Phospho-CDKN2A (Ser152) Polyclonal Antibody for Western Blot, IHC (P) and pep-ELISA
  • Host Rabbit
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # PA539802
  • Size 100µg
Applications
Immunohistochemistry (Paraffin) (1:50-1:100), peptide-ELISA (1:1000), Western Blot (1:500-1:1000)
Alternate names:
p14ARF; Cyclin-dependent kinase inhibitor 2A; Alternative reading frame; p16-INK4a; Multiple tumor suppressor 1; Cyclin-dependent kinase 4 inhibitor A
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA539802 309.00
Check your price
  • CDKN2A Monoclonal Antibody for Western Blot, IHC and Flow
  • Host Mouse
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # MA517054
  • Size 100µL
Applications
Flow Cytometry (1:200-1:400), Immunohistochemistry (1:200-1:1000), Western Blot (1:500-1:2000)
Alternate names:
p14ARF; Cyclin-dependent kinase inhibitor 2A; Alternative reading frame; p16-INK4a; Multiple tumor suppressor 1; Cyclin-dependent kinase 4 inhibitor A
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA517054 309.00
Check your price
  • CDKN2A Polyclonal Antibody for IHC (P) and IP
  • Host Rabbit
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # PA532529
  • Size 500µL
Applications
Immunohistochemistry (Paraffin) (1:25)
Alternate names:
p14ARF; Cyclin-dependent kinase inhibitor 2A; Alternative reading frame; p16-INK4a; Multiple tumor suppressor 1; Cyclin-dependent kinase 4 inhibitor A
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA532529 275.00
Check your price
  • CDKN2C Polyclonal Antibody for Western Blot and IHC
  • Host Rabbit
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # PA524435
  • Size 400µL
Applications
Immunohistochemistry (1:50-1:100), Western Blot (1:1000)
Alternate names:
p18-INK6; p18-INK4c; Cyclin-dependent kinase 6 inhibitor
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA524435 349.00
Check your price
  • CDKN2B Monoclonal Antibody for IF, ICC and IP
  • Host Mouse
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # MA191096
  • Size 500µL
Applications
Immunocytochemistry (Assay-dependent), Immunofluorescence (Assay dependent), Immunoprecipitation (1:400)
Alternate names:
p15-INK4b; p14-INK4b; Multiple tumor suppressor 2
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA191096 349.00
Check your price
  • CDKN2B Polyclonal Antibody for Western Blot
  • Host Rabbit
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # PA124258
  • Size 100µg
Applications
Western Blot (1 µg/ml)
Alternate names:
p15-INK4b; p14-INK4b; Multiple tumor suppressor 2
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA124258 399.00
Check your price
  • Phospho-CDKN2A (Ser326) Polyclonal Antibody for Western Blot
  • Host Rabbit
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # PA540273
  • Size 100µl
Applications
Western Blot (1:500-1:2000)
Alternate names:
p14ARF; Cyclin-dependent kinase inhibitor 2A; Alternative reading frame; p16-INK4a; Multiple tumor suppressor 1; Cyclin-dependent kinase 4 inhibitor A
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA540273 309.00
Check your price
  • CDKN2A Polyclonal Antibody for IHC (P) and IP
  • Host Rabbit
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # PA138334
  • Size 1mL
Applications
Immunohistochemistry (Paraffin) (1:25)
Alternate names:
p14ARF; Cyclin-dependent kinase inhibitor 2A; Alternative reading frame; p16-INK4a; Multiple tumor suppressor 1; Cyclin-dependent kinase 4 inhibitor A
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA138334 399.00
Check your price
  • CDKN2A Polyclonal Antibody for Western Blot, IHC, IHC (F), IHC (P) and ELISA
  • Host Rabbit
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # PA116661
  • Size 100µg
Applications
ELISA (1:100-1:2000), Immunohistochemistry (Frozen) (1:500-1:1000), Immunohistochemistry (Paraffin) (1:500-1:1000), Western Blot (1:500-1:2500)
Alternate names:
p14ARF; Cyclin-dependent kinase inhibitor 2A; Alternative reading frame; p16-INK4a; Multiple tumor suppressor 1; Cyclin-dependent kinase 4 inhibitor A
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA116661 349.00
Check your price
  • CDKN2A Polyclonal Antibody for Western Blot, IHC, IHC (P) and ELISA
  • Host Rabbit
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # PA116662
  • Size 100µg
Applications
ELISA (1:100-1:2000), Immunohistochemistry (Paraffin) (1:100-1:500), Western Blot (1:500-1:2500)
Alternate names:
p14ARF; Cyclin-dependent kinase inhibitor 2A; Alternative reading frame; p16-INK4a; Multiple tumor suppressor 1; Cyclin-dependent kinase 4 inhibitor A
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA116662 325.00
Check your price