1 - 30 of 30 products displayed
  • hCG beta Polyclonal Antibody for IHC (F) and IHC (P)
  • Host Rabbit
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # PA127113
  • Size 500 µL
Applications
Immunohistochemistry (Frozen) (1:100-1:500), Immunohistochemistry (Paraffin) (1:100-1:500)
Alternate names:
Chorionic gonadotrophin chain beta
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA127113 309.00
Check your price
  • hCG beta Polyclonal Antibody for IHC (F) and IHC (P)
  • Host Rabbit
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # PA126300
  • Size 6 mL
Applications
Immunohistochemistry (Frozen) (Neat), Immunohistochemistry (Paraffin) (Neat)
Alternate names:
Chorionic gonadotrophin chain beta
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA126300 375.00
Check your price
  • hCG beta Monoclonal Antibody, HRP conjugate for ELISA
  • Host Mouse
  • Target Species Human
  • Conjugate HRP
  • Catalog # MA121651
  • Size 200 µg
Applications
ELISA (Assay dependent)
Alternate names:
Chorionic gonadotrophin chain beta
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA121651 349.00
Check your price
  • hCG beta Monoclonal Antibody for ELISA
  • Host Mouse
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # MA121656
  • Size 500 µg
Applications
ELISA (Assay dependent)
Alternate names:
Chorionic gonadotrophin chain beta
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA121656 309.00
Check your price
  • hCG beta Monoclonal Antibody for ELISA
  • Host Mouse
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # MA121654
  • Size 500 µg
Applications
ELISA (Assay dependent)
Alternate names:
Chorionic gonadotrophin chain beta
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA121654 309.00
Check your price
  • hCG beta-4 Monoclonal Antibody for ELISA and RIA
  • Host Mouse
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # MA125110
  • Size 250 µg
Applications
ELISA (Assay Dependent), Radioimmune Assays (Assay Dependent)
Alternate names:
Chorionic gonadotrophin chain beta
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA125110 349.00
Check your price
  • hCG alpha Polyclonal Antibody for ELISA
  • Host Goat
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # PA127276
  • Size 100 µg
Applications
ELISA (Assay Dependent)
Alternate names:
Thyrotropin alpha chain; Thyroid-stimulating hormone alpha chain; Lutropin alpha chain; Luteinizing hormone alpha chain; Follitropin alpha chain; Follicle-stimulating hormone alpha chain; Chorionic gonadotrophin subunit alpha; Choriogonadotropin alpha chain; Anterior pituitary glycoprotein hormones common subunit alpha
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA127276 349.00
Check your price
  • hCG Monoclonal Antibody for IP, ELISA and RIA
  • Host Mouse
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # MA514751
  • Size 100 ug
Applications
ELISA (Assay Dependent), Immunoprecipitation (Assay Dependent), Radioimmune Assays (Assay Dependent)
Alternate names:
Chorionic gonadotrophin chain beta
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA514751 349.00
Check your price
  • hCG Monoclonal Antibody for Western Blot, IP, ELISA and RIA
  • Host Mouse
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # MA514702
  • Size 100 ug
Applications
ELISA (Assay Dependent), Immunoprecipitation (Assay Dependent), Radioimmune Assays (Assay Dependent), Western Blot (1:250-1:500)
Alternate names:
Chorionic gonadotrophin chain beta
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA514702 349.00
Check your price
  • hCG Monoclonal Antibody for IHC (P), IP, ELISA and RIA
  • Host Mouse
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # MA514699
  • Size 100 ug
Applications
ELISA (2-10µg/ml), Immunohistochemistry (Paraffin) (1:20), Immunoprecipitation (5µg/ml), Radioimmune Assays (Assay Dependent)
Alternate names:
Chorionic gonadotrophin chain beta
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA514699 349.00
Check your price
  • hCG Monoclonal Antibody for Western Blot, IHC (P), IP, ELISA and RIA
  • Host Mouse
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # MA514700
  • Size 100 ug
Applications
ELISA (Assay Dependent), Immunohistochemistry (Paraffin) (1:20), Immunoprecipitation (Assay Dependent), Radioimmune Assays (Assay Dependent), Western Blot (1:250-1:500)
Alternate names:
Chorionic gonadotrophin chain beta
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA514700 349.00
Check your price
  • hCG Monoclonal Antibody for Western Blot, IHC (P), IP, ELISA and RIA
  • Host Mouse
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # MA514680
  • Size 100 ug
Applications
ELISA (2-10µg/ml), Immunohistochemistry (Paraffin) (1:20), Immunoprecipitation (5µg/ml), Radioimmune Assays (Assay Dependent), Western Blot (1:250-1:500)
Alternate names:
Chorionic gonadotrophin chain beta
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA514680 349.00
Check your price
  • hCG beta Polyclonal Antibody for Western Blot
  • Host Chicken
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # PA127846
  • Size 50 µg
Applications
Western Blot (Assay Dependent)
Alternate names:
Chorionic gonadotrophin chain beta
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA127846 425.00
Check your price
  • hCG Monoclonal Antibody for IP, ELISA and RIA
  • Host Mouse
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # MIH9820
  • Size 1 mg
Applications
ELISA (Assay Dependent), Immunoprecipitation (Assay Dependent), Radioimmune Assays (Assay Dependent)
Alternate names:
Chorionic gonadotrophin chain beta
Catalog # Size Price (AUD) Your Price (AUD) Qty
MIH9820 600.00
Check your price
  • hCG Monoclonal Antibody for Western Blot, IP, ELISA and RIA
  • Host Mouse
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # MIH9822
  • Size 1 mg
Applications
ELISA (Assay Dependent), Immunoprecipitation (Assay Dependent), Radioimmune Assays (Assay Dependent), Western Blot (1:250-1:500)
Alternate names:
Chorionic gonadotrophin chain beta
Catalog # Size Price (AUD) Your Price (AUD) Qty
MIH9822 600.00
Check your price
  • hCG Monoclonal Antibody for IHC (P), IP, ELISA and RIA
  • Host Mouse
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # MIH9801
  • Size 1 mg
Applications
ELISA (2-10µg/ml), Immunohistochemistry (Paraffin) (1:20), Immunoprecipitation (5µg/ml), Radioimmune Assays (Assay Dependent)
Alternate names:
Chorionic gonadotrophin chain beta
Catalog # Size Price (AUD) Your Price (AUD) Qty
MIH9801 600.00
Check your price
  • hCG Monoclonal Antibody for Western Blot, IHC (P), IP, ELISA and RIA
  • Host Mouse
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # MIH9805
  • Size 1 mg
Applications
ELISA (Assay Dependent), Immunohistochemistry (Paraffin) (1:20), Immunoprecipitation (Assay Dependent), Radioimmune Assays (Assay Dependent), Western Blot (1:250-1:500)
Alternate names:
Chorionic gonadotrophin chain beta
Catalog # Size Price (AUD) Your Price (AUD) Qty
MIH9805 600.00
Check your price
  • hCG Monoclonal Antibody for Western Blot, IP, ELISA and RIA
  • Host Mouse
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # MIH9813
  • Size 1 mg
Applications
ELISA (Assay Dependent), Immunoprecipitation (5µg/ml), Radioimmune Assays (Assay Dependent), Western Blot (1:250-1:500)
Alternate names:
Chorionic gonadotrophin chain beta
Catalog # Size Price (AUD) Your Price (AUD) Qty
MIH9813 600.00
Check your price
  • hCG Monoclonal Antibody for Western Blot, IP, ELISA and RIA
  • Host Mouse
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # MA514701
  • Size 100 ug
Applications
ELISA (Assay Dependent), Immunoprecipitation (5µg/ml), Radioimmune Assays (Assay Dependent), Western Blot (1:250-1:500)
Alternate names:
Chorionic gonadotrophin chain beta
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA514701 349.00
Check your price
  • hCG Monoclonal Antibody for Western Blot, IHC (P), IP, ELISA and RIA
  • Host Mouse
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # MIH9812
  • Size 1 mg
Applications
ELISA (2-10µg/ml), Immunohistochemistry (Paraffin) (1:20), Immunoprecipitation (5µg/ml), Radioimmune Assays (Assay Dependent), Western Blot (1:250-1:500)
Alternate names:
Chorionic gonadotrophin chain beta
Catalog # Size Price (AUD) Your Price (AUD) Qty
MIH9812 600.00
Check your price
  • hCG beta Monoclonal Antibody for ICC and ELISA
  • Host Mouse
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # MA119065
  • Size 100 µg
Applications
ELISA (Assay Dependent), Immunocytochemistry (Assay Dependent)
Alternate names:
Chorionic gonadotrophin chain beta
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA119065 159.00
Check your price
  • hCG beta Monoclonal Antibody for ICC and ELISA
  • Host Mouse
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # MA119063
  • Size 100 µg
Applications
ELISA (Assay Dependent), Immunocytochemistry (Assay Dependent)
Alternate names:
Chorionic gonadotrophin chain beta
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA119063 159.00
Check your price
  • Thyroid Stimulating Hormone Monoclonal Antibody for IHC (P)
  • Host Mouse
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # MA512159
  • Size 500 µL
Applications
Immunohistochemistry (Paraffin) (1:200-1:400)
Alternate names:
Thyrotropin alpha chain; Thyroid-stimulating hormone alpha chain; Lutropin alpha chain; Luteinizing hormone alpha chain; Follitropin alpha chain; Follicle-stimulating hormone alpha chain; Chorionic gonadotrophin subunit alpha; Choriogonadotropin alpha chain; Anterior pituitary glycoprotein hormones common subunit alpha
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA512159 225.00
Check your price
  • hCG alpha-4 Monoclonal Antibody for Western Blot, IHC (P), IP, ELISA and RIA
  • Host Mouse
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # MA125038
  • Size 250 µg
Applications
ELISA (Assay Dependent), Immunohistochemistry (Paraffin) (Assay Dependent), Immunoprecipitation (Assay Dependent), Radioimmune Assays (Assay Dependent), Western Blot (Assay-dependent)
Alternate names:
Thyrotropin alpha chain; Thyroid-stimulating hormone alpha chain; Lutropin alpha chain; Luteinizing hormone alpha chain; Follitropin alpha chain; Follicle-stimulating hormone alpha chain; Chorionic gonadotrophin subunit alpha; Choriogonadotropin alpha chain; Anterior pituitary glycoprotein hormones common subunit alpha
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA125038 349.00
Check your price
  • hCG alpha-6 Monoclonal Antibody for Western Blot, ELISA and RIA
  • Host Mouse
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # MA125108
  • Size 250 µg
Applications
ELISA (Assay Dependent), Radioimmune Assays (Assay Dependent), Western Blot (Assay-dependent)
Alternate names:
Thyrotropin alpha chain; Thyroid-stimulating hormone alpha chain; Lutropin alpha chain; Luteinizing hormone alpha chain; Follitropin alpha chain; Follicle-stimulating hormone alpha chain; Chorionic gonadotrophin subunit alpha; Choriogonadotropin alpha chain; Anterior pituitary glycoprotein hormones common subunit alpha
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA125108 349.00
Check your price
  • hCG alpha Monoclonal Antibody for Western Blot and ELISA
  • Host Mouse
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # MA127124
  • Size 500 µg
Applications
ELISA (Assay Dependent), Western Blot (Assay Dependent)
Alternate names:
Thyrotropin alpha chain; Thyroid-stimulating hormone alpha chain; Lutropin alpha chain; Luteinizing hormone alpha chain; Follitropin alpha chain; Follicle-stimulating hormone alpha chain; Chorionic gonadotrophin subunit alpha; Choriogonadotropin alpha chain; Anterior pituitary glycoprotein hormones common subunit alpha
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA127124 349.00
Check your price
  • hCG alpha Monoclonal Antibody for IP, ELISA and RIA
  • Host Mouse
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # MA121405
  • Size 500 µg
Applications
ELISA (Assay dependent), Immunoprecipitation (Assay dependent), Radioimmune Assays (Assay-dependent)
Alternate names:
Thyrotropin alpha chain; Thyroid-stimulating hormone alpha chain; Lutropin alpha chain; Luteinizing hormone alpha chain; Follitropin alpha chain; Follicle-stimulating hormone alpha chain; Chorionic gonadotrophin subunit alpha; Choriogonadotropin alpha chain; Anterior pituitary glycoprotein hormones common subunit alpha
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA121405 399.00
Check your price
  • hCG beta Monoclonal Antibody for IP, ELISA and RIA
  • Host Mouse
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # MA514703
  • Size 100 ug
Applications
ELISA (2-10µg/ml), Immunoprecipitation (Assay Dependent), Radioimmune Assays (Assay Dependent)
Alternate names:
Chorionic gonadotrophin chain beta
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA514703 349.00
Check your price
  • hCG beta Monoclonal Antibody for ELISA and RIA
  • Host Mouse
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # MA518033
  • Size 100 µL
Applications
ELISA (Assay-dependent), Radioimmune Assays (Assay-dependent)
Alternate names:
Chorionic gonadotrophin chain beta
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA518033 399.00
Check your price
  • hCG beta Monoclonal Antibody for IP, ELISA and RIA
  • Host Mouse
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # MIH9827
  • Size 1 mg
Applications
ELISA (2-10µg/ml), Immunoprecipitation (Assay Dependent), Radioimmune Assays (Assay Dependent)
Alternate names:
Chorionic gonadotrophin chain beta
Catalog # Size Price (AUD) Your Price (AUD) Qty
MIH9827 600.00
Check your price