1 - 30 of 30 products displayed
  • Growth Hormone Monoclonal Antibody for Western Blot and IHC (P)
  • Host Mouse
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # MA511926
  • Size 500 µL
Applications
Immunohistochemistry (Paraffin) (1:2000-1:4000), Western Blot (1-2 µg/ml)
Alternate names:
Pituitary growth hormone; Growth hormone 1; Growth hormone
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA511926 309.00
Check your price
  • Growth Hormone Polyclonal Antibody for Western Blot and IHC (P)
  • Host Rabbit
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # PA527360
  • Size 100 µL
Applications
Immunohistochemistry (Paraffin) (1:100-1:1000), Western Blot (1:500-1:3000)
Alternate names:
Pituitary growth hormone; Growth hormone 1; Growth hormone
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA527360 349.00
Check your price
  • Growth Hormone Polyclonal Antibody for IHC (P)
  • Host Rabbit
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # PA532448
  • Size 500 µL
Applications
Immunohistochemistry (Paraffin) (1:200)
Alternate names:
Pituitary growth hormone; Growth hormone 1; Growth hormone
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA532448 349.00
Check your price
  • Growth Hormone Polyclonal Antibody for IHC (F) and IHC (P)
  • Host Rabbit
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # PA136094
  • Size 250 µL
Applications
Immunohistochemistry (Paraffin, Frozen) (1:50)
Alternate names:
Pituitary growth hormone; Growth hormone 1; Growth hormone
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA136094 425.00
Check your price
  • Growth Hormone Monoclonal Antibody for IP, ELISA and RIA
  • Host Mouse
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # MA514682
  • Size 100 ug
Applications
ELISA (Assay Dependent), Immunoprecipitation (Assay Dependent), Radioimmune Assays (Assay Dependent)
Alternate names:
Pituitary growth hormone; Growth hormone 1; Growth hormone
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA514682 349.00
Check your price
  • Growth Hormone Monoclonal Antibody for IP, ELISA and RIA
  • Host Mouse
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # MIH9901
  • Size 1 mg
Applications
ELISA (Assay Dependent), Immunoprecipitation (Assay Dependent), Radioimmune Assays (Assay Dependent)
Alternate names:
Pituitary growth hormone; Growth hormone 1; Growth hormone
Catalog # Size Price (AUD) Your Price (AUD) Qty
MIH9901 600.00
Check your price
  • Growth Hormone Polyclonal Antibody for IHC (P)
  • Host Rabbit
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # PA133080
  • Size 6 mL
Applications
Immunohistochemistry (Paraffin) (Neat)
Alternate names:
Pituitary growth hormone; Growth hormone 1; Growth hormone
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA133080 425.00
Check your price
  • Growth Hormone Polyclonal Antibody for ELISA and RIA
  • Host Sheep
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # PA185519
  • Size 250 µL
Applications
ELISA (Assay Dependent), Radioimmune Assays (Assay Dependent)
Alternate names:
Pituitary growth hormone; Growth hormone 1; Growth hormone
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA185519 249.00
Check your price
  • Growth Hormone Receptor Polyclonal Antibody for Western Blot, IHC, Flow and Neu
  • Host Goat
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # PA547094
  • Size 100 µg
Applications
Flow Cytometry (2.5 µg/10^6 cells), Immunohistochemistry (5-15 µg/ml), Neutralization (0.125-0.5 µg/ml), Western Blot (0.1 µg/ml)
Alternate names:
Somatotropin receptor
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA547094 349.00
Check your price
  • Growth Hormone Polyclonal Antibody for Western Blot, IF, IHC (F) and IHC (P)
  • Host Rabbit
  • Target Species Dog, Cat, Human, Sheep, Rat
  • Conjugate Unconjugated
  • Catalog # PA185518
  • Size 50 µL
Applications
Immunofluorescence (1/10-1/100), Immunohistochemistry (Frozen) (Assay Dependent), Immunohistochemistry (Paraffin) (1/500-1/4000), Western Blot (1/50-1/500)
Alternate names:
Growth hormone; Pituitary growth hormone; Growth hormone 1
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA185518 349.00
Check your price
  • Growth Hormone Polyclonal Antibody for Western Blot and IHC (F)
  • Host Goat
  • Target Species Rat
  • Conjugate Unconjugated
  • Catalog # PA546911
  • Size 100 µg
Applications
Immunohistochemistry (Frozen) (5-15 µg/ml), Western Blot (0.1 µg/ml)
Alternate names:
Growth hormone
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA546911 375.00
Check your price
  • Growth Hormone Receptor Polyclonal Antibody for Western Blot, IHC and Neu
  • Host Goat
  • Target Species Rat
  • Conjugate Unconjugated
  • Catalog # PA547095
  • Size 100 µg
Applications
Immunohistochemistry (5-15 µg/ml), Neutralization (0.075-0.3 µg/ml), Western Blot (0.1 µg/ml)
Alternate names:
Somatotropin receptor
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA547095 349.00
Check your price
  • Growth Hormone Receptor Polyclonal Antibody for Western Blot and IHC (F)
  • Host Goat
  • Target Species Mouse
  • Conjugate Unconjugated
  • Catalog # PA547128
  • Size 100 µg
Applications
Immunohistochemistry (Frozen) (5-15 µg/ml), Western Blot (0.1 µg/ml)
Alternate names:
Somatotropin receptor
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA547128 349.00
Check your price
  • Growth Hormone Monoclonal Antibody for ELISA
  • Host Mouse
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # MA121649
  • Size 500 µg
Applications
ELISA (Assay dependent)
Alternate names:
Pituitary growth hormone; Growth hormone 1; Growth hormone
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA121649 309.00
Check your price
  • Growth Hormone Monoclonal Antibody for ELISA
  • Host Mouse
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # MA121650
  • Size 500 µg
Applications
ELISA (Assay dependent)
Alternate names:
Pituitary growth hormone; Growth hormone 1; Growth hormone
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA121650 309.00
Check your price
  • Growth Hormone Monoclonal Antibody for ELISA
  • Host Mouse
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # MA121648
  • Size 500 µg
Applications
ELISA (Assay dependent)
Alternate names:
Pituitary growth hormone; Growth hormone 1; Growth hormone
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA121648 309.00
Check your price
  • Growth Hormone Polyclonal Antibody for Western Blot and ELISA
  • Host Chicken
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # PA127501
  • Size 50 µg
Applications
ELISA (0.1 µg/ml), Western Blot (1:2,000)
Alternate names:
Pituitary growth hormone; Growth hormone 1; Growth hormone
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA127501 425.00
Check your price
  • Growth Hormone Receptor Polyclonal Antibody for Western Blot
  • Host Rabbit
  • Target Species Dog, Horse, Human, Pig
  • Conjugate Unconjugated
  • Catalog # PA542544
  • Size 100 µl
Applications
Western Blot (1 µg/ml )
Alternate names:
Somatotropin receptor
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA542544 309.00
Check your price
  • Growth Hormone Receptor Polyclonal Antibody for Western Blot
  • Host Rabbit
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # PA542542
  • Size 100 µl
Applications
Western Blot (1 µg/ml )
Alternate names:
Somatotropin receptor
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA542542 309.00
Check your price
  • Growth Hormone Receptor Polyclonal Antibody for Western Blot
  • Host Rabbit
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # PA542543
  • Size 100 µl
Applications
Western Blot (0.2-1 µg/ml)
Alternate names:
Somatotropin receptor
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA542543 309.00
Check your price
  • GH2 Polyclonal Antibody for Western Blot
  • Host Rabbit
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # PA534767
  • Size 100 µL
Applications
Western Blot (1:1000-1:10,000)
Alternate names:
Placenta-specific growth hormone; Growth hormone 2
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA534767 349.00
Check your price
  • Human Growth Hormone for Ctrl
  • Host E. Coli
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # RP10928
  • Size 1000 µg
Applications
Control (Assay Dependent)
Alternate names:
HGH
Catalog # Size Price (AUD) Your Price (AUD) Qty
RP10928 412.00
Check your price
  • Human Growth Hormone for Ctrl
  • Host E. coli
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # RP10912
  • Size 500 µg
Applications
Control (Assay Dependent)
Alternate names:
HGH
Catalog # Size Price (AUD) Your Price (AUD) Qty
RP10912 307.00
Check your price
  • Thyroid Stimulating Hormone Monoclonal Antibody for IHC (P)
  • Host Mouse
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # MA512159
  • Size 500 µL
Applications
Immunohistochemistry (Paraffin) (1:200-1:400)
Alternate names:
Thyrotropin alpha chain; Thyroid-stimulating hormone alpha chain; Lutropin alpha chain; Luteinizing hormone alpha chain; Follitropin alpha chain; Follicle-stimulating hormone alpha chain; Chorionic gonadotrophin subunit alpha; Choriogonadotropin alpha chain; Anterior pituitary glycoprotein hormones common subunit alpha
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA512159 225.00
Check your price
  • Follicle Stimulating Hormone Monoclonal Antibody for IHC (P)
  • Host Mouse
  • Target Species Bovine, Human
  • Conjugate Unconjugated
  • Catalog # MA512144
  • Size 500 µL
Applications
Immunohistochemistry (Paraffin) (0.2-0.4 µg/mL)
Alternate names:
Follitropin beta chain; Follicle-stimulating hormone beta subunit
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA512144 225.00
Check your price
  • Follicle Stimulating Hormone Monoclonal Antibody for IHC (F), IHC (P) and ELISA
  • Host Mouse
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # MA124935
  • Size 250 µg
Applications
ELISA (Assay Dependent), Immunohistochemistry (Frozen) (Assay Dependent), Immunohistochemistry (Paraffin) (Assay Dependent)
Alternate names:
Follitropin beta chain; Follicle-stimulating hormone beta subunit
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA124935 399.00
Check your price
  • Prolactin Monoclonal Antibody for Western Blot, IHC and ELISA
  • Host Mouse
  • Target Species Bovine, Human, Mouse, Non-human primate, Sheep, Rat
  • Conjugate Unconjugated
  • Catalog # MA110597
  • Size 100 ug
Applications
ELISA (Assay dependent), Immunohistochemistry (Assay dependent), Western Blot (Assay dependent)
Alternate names:
Prl
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA110597 399.00
Check your price
  • GHRHR Polyclonal Antibody for IHC (P)
  • Host Rabbit
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # PA533583
  • Size 50 ug
Applications
Immunohistochemistry (Paraffin) (32 µg/ml)
Alternate names:
Growth hormone-releasing factor receptor
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA533583 349.00
Check your price
  • GHRHR Polyclonal Antibody for IHC (P)
  • Host Rabbit
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # PA533584
  • Size 50 ug
Applications
Immunohistochemistry (Paraffin) (10-16 µg/ml)
Alternate names:
Growth hormone-releasing factor receptor
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA533584 349.00
Check your price
  • GHRHR Polyclonal Antibody for IHC (P)
  • Host Rabbit
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # PA533582
  • Size 50 ug
Applications
Immunohistochemistry (Paraffin) (3 µg/ml)
Alternate names:
Growth hormone-releasing factor receptor
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA533582 349.00
Check your price