1 - 30 of 30 products displayed
  • CD79a Monoclonal Antibody for IHC (P) and Flow
  • Host Rabbit
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # MA516344
  • Size 500µL
Applications
Flow Cytometry (1:100), Immunohistochemistry (Paraffin) (1:400)
Alternate names:
Surface IgM-associated protein; Membrane-bound immunoglobulin-associated protein; MB-1 membrane glycoprotein; Ig-alpha
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA516344 375.00
Check your price
  • CD79a Monoclonal Antibody, Biotin conjugate for Western Blot, IF and IHC (P)
  • Host Mouse
  • Target Species Bovine, Chicken, Horse, Guinea pig, Human, Mouse, Non-human primate, Pig, Rabbit, Rat
  • Conjugate Biotin
  • Catalog # MA513209
  • Size 500µL
Applications
Immunofluorescence (Assay Dependent), Immunohistochemistry (Paraffin) (0.5-1 ug/ml), Western Blot (0.5-1.0 µg/ml)
Alternate names:
Surface IgM-associated protein; Membrane-bound immunoglobulin-associated protein; MB-1 membrane glycoprotein; Ig-alpha
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA513209 375.00
Check your price
  • CD79a Monoclonal Antibody for Western Blot, IF, ICC, IHC (P) and Flow
  • Host Mouse
  • Target Species Bovine, Chicken, Horse, Guinea pig, Human, Mouse, Non-human primate, Pig, Rabbit, Rat
  • Conjugate Unconjugated
  • Catalog # MA513212
  • Size 500µL
Applications
Immunofluorescence (Assay Dependent), Immunohistochemistry (Paraffin) (0.5-1.0 µg/ml), Western Blot (0.5-1.0 µg/ml)
Alternate names:
Surface IgM-associated protein; Membrane-bound immunoglobulin-associated protein; MB-1 membrane glycoprotein; Ig-alpha
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA513212 309.00
Check your price
  • CD79a Monoclonal Antibody for Western Blot, IHC (P) and Flow
  • Host Rabbit
  • Target Species Human, Mouse
  • Conjugate Unconjugated
  • Catalog # MA514556
  • Size 500µL
Applications
Flow Cytometry (1:2), Immunohistochemistry (Paraffin) (1:100-1:300), Western Blot (1:1000-1:10,000)
Alternate names:
Surface IgM-associated protein; Membrane-bound immunoglobulin-associated protein; MB-1 membrane glycoprotein; Ig-alpha
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA514556 349.00
Check your price
  • CD79a Monoclonal Antibody for IHC (P) and Flow
  • Host Rabbit
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # MA139569
  • Size 7mL
Applications
Flow Cytometry (1:100), Immunohistochemistry (Paraffin) (Ready-to-use)
Alternate names:
Surface IgM-associated protein; Membrane-bound immunoglobulin-associated protein; MB-1 membrane glycoprotein; Ig-alpha
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA139569 249.00
Check your price
  • CD79a Polyclonal Antibody for Western Blot and IHC (P)
  • Host Rabbit
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # PA532333
  • Size 500µL
Applications
Immunohistochemistry (Paraffin) (1:200), Western Blot (1:25)
Alternate names:
Surface IgM-associated protein; Membrane-bound immunoglobulin-associated protein; MB-1 membrane glycoprotein; Ig-alpha
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA532333 225.00
Check your price
  • CD79a Monoclonal Antibody for Western Blot, IF, ICC, IHC (P) and IP
  • Host Mouse
  • Target Species Cat, Human, Mouse, Rabbit, Rat
  • Conjugate Unconjugated
  • Catalog # MA515234
  • Size 500µL
Applications
Immunofluorescence (Assay Dependent), Immunohistochemistry (Paraffin) (1:50-1:100), Immunoprecipitation (10 µg/ml), Western Blot (1:50)
Alternate names:
Surface IgM-associated protein; Membrane-bound immunoglobulin-associated protein; MB-1 membrane glycoprotein; Ig-alpha
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA515234 349.00
Check your price
  • CD79a Monoclonal Antibody, PE conjugate for Flow
  • Host Mouse
  • Target Species Human
  • Conjugate PE
  • Catalog # MHCD79a04
  • Size 0.5ml
Applications
Flow Cytometry (Assay-Dependent)
Alternate names:
Surface IgM-associated protein; Membrane-bound immunoglobulin-associated protein; MB-1 membrane glycoprotein; Ig-alpha
Catalog # Size Price (AUD) Your Price (AUD) Qty
MHCD79a04 Not Orderable Online
Check your price
  • CD79a Monoclonal Antibody for Flow
  • Host Mouse
  • Target Species Human
  • Conjugate
  • Catalog # A14946
  • Size 25tests
Applications
Flow Cytometry (Assay Dependent)
Alternate names:
Surface IgM-associated protein; Membrane-bound immunoglobulin-associated protein; MB-1 membrane glycoprotein; Ig-alpha
Catalog # Size Price (AUD) Your Price (AUD) Qty
A14946 Not Orderable Online
Check your price
  • CD79a Polyclonal Antibody for Western Blot, IF and ICC
  • Host Rabbit
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # A16575
  • Size 100µL
Applications
Immunocytochemistry (Assay Dependent), Immunofluorescence (Assay Dependent), Western Blot (Assay Dependent)
Alternate names:
Surface IgM-associated protein; Membrane-bound immunoglobulin-associated protein; MB-1 membrane glycoprotein; Ig-alpha
Catalog # Size Price (AUD) Your Price (AUD) Qty
A16575 309.00
Check your price
  • CD79a Monoclonal Antibody for Western Blot, IHC (P) and Flow
  • Host Mouse
  • Target Species Human, Mouse
  • Conjugate Unconjugated
  • Catalog # MA511636
  • Size 500µL
Applications
Flow Cytometry (2 ug/test), Immunohistochemistry (Paraffin) (1:50-1:100), Western Blot (1:100-1:500)
Alternate names:
Surface IgM-associated protein; Membrane-bound immunoglobulin-associated protein; MB-1 membrane glycoprotein; Ig-alpha
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA511636 475.00
Check your price
  • CD79a Monoclonal Antibody for Western Blot, IHC (F), IHC (P) and Flow
  • Host Mouse
  • Target Species Many
  • Conjugate Unconjugated
  • Catalog # MA516710
  • Size 200ug
Applications
Flow Cytometry (1:10-1:50), Immunohistochemistry (Frozen) (Assay Dependent), Immunohistochemistry (Paraffin) (1:100-1:500), Western Blot (Assay Dependent)
Alternate names:
Surface IgM-associated protein; Membrane-bound immunoglobulin-associated protein; MB-1 membrane glycoprotein; Ig-alpha
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA516710 375.00
Check your price
  • CD79a Monoclonal Antibody for IHC (F), IHC (P) and Flow
  • Host Mouse
  • Target Species Many
  • Conjugate Unconjugated
  • Catalog # MA181870
  • Size 100µg
Applications
Flow Cytometry (1:50-1:100), Immunohistochemistry (Frozen) (Assay Dependent), Immunohistochemistry (Paraffin) (1:100-1:500)
Alternate names:
Surface IgM-associated protein; Membrane-bound immunoglobulin-associated protein; MB-1 membrane glycoprotein; Ig-alpha
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA181870 309.00
Check your price
  • CD79a Monoclonal Antibody, PE conjugate for Flow
  • Host Mouse
  • Target Species Many
  • Conjugate PE
  • Catalog # MA516711
  • Size 100TESTS
Applications
Flow Cytometry (Neat)
Alternate names:
Surface IgM-associated protein; Membrane-bound immunoglobulin-associated protein; MB-1 membrane glycoprotein; Ig-alpha
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA516711 425.00
Check your price
  • Mouse anti-Human IgA Secondary Antibody for IHC (P) and ELISA
  • Host Mouse
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # SA119258
  • Size 100µg
Applications
ELISA (Assay Dependent), Immunohistochemistry (Paraffin) (10µg/ml)
Alternate names:
Catalog # Size Price (AUD) Your Price (AUD) Qty
SA119258 137.00
Check your price
  • Goat anti-Human IgA Secondary Antibody, Biotin conjugate for IF, Flow and ELISA
  • Host Goat
  • Target Species Human
  • Conjugate Biotin
  • Catalog # A18785
  • Size 1mg
Applications
ELISA (1:500-1:20,000), Flow Cytometry (1:500-1:20,000), Immunofluorescence (1:500-1:20,000)
Alternate names:
Catalog # Size Price (AUD) Your Price (AUD) Qty
A18785 106.00
Check your price
  • Goat anti-Human IgA Secondary Antibody, TRITC conjugate for IF
  • Host Goat
  • Target Species Human
  • Conjugate TRITC
  • Catalog # A18786
  • Size 1mg
Applications
Immunofluorescence (1:500-1:20,000)
Alternate names:
Catalog # Size Price (AUD) Your Price (AUD) Qty
A18786 96.00
Check your price
  • Goat anti-Human IgA Secondary Antibody, FITC conjugate for IF and Flow
  • Host Goat
  • Target Species Human
  • Conjugate FITC
  • Catalog # A18782
  • Size 1mg
Applications
Flow Cytometry (1:500-1:20,000), Immunofluorescence (1:500-1:20,000)
Alternate names:
Catalog # Size Price (AUD) Your Price (AUD) Qty
A18782 94.00
Check your price
  • Goat anti-Human IgA Secondary Antibody, HRP conjugate for Western Blot, IHC and ELISA
  • Host Goat
  • Target Species Human
  • Conjugate HRP
  • Catalog # A18781
  • Size 1mg
Applications
ELISA (1:500-1:20,000), Immunohistochemistry (1:500-1:20,000), Western Blot (1:500-1:20,000)
Alternate names:
Catalog # Size Price (AUD) Your Price (AUD) Qty
A18781 109.00
Check your price
  • Goat anti-Human IgA Secondary Antibody, AP conjugate for Western Blot, IHC and ELISA
  • Host Goat
  • Target Species Human
  • Conjugate AP
  • Catalog # A18784
  • Size 1mg
Applications
ELISA (1:500-1:20,000), Immunohistochemistry (1:500-1:20,000), Western Blot (1:500-1:20,000)
Alternate names:
Catalog # Size Price (AUD) Your Price (AUD) Qty
A18784 157.00
Check your price
  • Goat anti-Human IgA Secondary Antibody for ELISA
  • Host Goat
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # A18783
  • Size 2mg
Applications
ELISA (1:500-1:20,000)
Alternate names:
Catalog # Size Price (AUD) Your Price (AUD) Qty
A18783 84.00
Check your price
  • Mouse anti-Human IgA Secondary Antibody, HRP conjugate for IHC (F), ELISA, RIA and DB
  • Host Mouse
  • Target Species Human
  • Conjugate HRP
  • Catalog # SA135467
  • Size 1mL
Applications
Dot blot (1:100), ELISA (1:1000), Immunohistochemistry (Frozen) (1:40), Radioimmune Assays (Assay dependent)
Alternate names:
Catalog # Size Price (AUD) Your Price (AUD) Qty
SA135467 886.00
Check your price
  • Goat anti-Human IgA Secondary Antibody, FITC conjugate for Western Blot, IF, IHC (P), IP and ELISA
  • Host Goat
  • Target Species Human
  • Conjugate FITC
  • Catalog # 627411
  • Size 1mL
Applications
ELISA (1:20-1:50), Immunofluorescence (1:20-1:50), Immunohistochemistry (Paraffin) (1:20-1:50), Immunoprecipitation (1:20-1:50), Western Blot (1:20-1:50)
Alternate names:
Catalog # Size Price (AUD) Your Price (AUD) Qty
627411 139.00
Check your price
  • F(ab')2-Goat anti-Human IgA Secondary Antibody, Biotin conjugate for IF, Flow and ELISA
  • Host Goat
  • Target Species Human
  • Conjugate Biotin
  • Catalog # A24462
  • Size 500µg
Applications
ELISA (1:500-1:20,000), Flow Cytometry (1:500-1:20,000), Immunofluorescence (1:500-1:20,000)
Alternate names:
Catalog # Size Price (AUD) Your Price (AUD) Qty
A24462 169.00
Check your price
  • F(ab')2-Goat anti-Human IgA Secondary Antibody, TRITC conjugate for IF
  • Host Goat
  • Target Species Human
  • Conjugate TRITC
  • Catalog # A24463
  • Size 500µg
Applications
Immunofluorescence (1:500-1:20,000)
Alternate names:
Catalog # Size Price (AUD) Your Price (AUD) Qty
A24463 150.00
Check your price
  • F(ab')2-Goat anti-Human IgA Secondary Antibody, FITC conjugate for IF and Flow
  • Host Goat
  • Target Species Human
  • Conjugate FITC
  • Catalog # A24459
  • Size 500µg
Applications
Flow Cytometry (1:500-1:20,000), Immunofluorescence (1:500-1:20,000)
Alternate names:
Catalog # Size Price (AUD) Your Price (AUD) Qty
A24459 150.00
Check your price
  • F(ab')2-Goat anti-Human IgA Secondary Antibody, HRP conjugate for Western Blot, IHC and ELISA
  • Host Goat
  • Target Species Human
  • Conjugate HRP
  • Catalog # A24458
  • Size 500µg
Applications
ELISA (1:500-1:20,000), Immunohistochemistry (1:500-1:20,000), Western Blot (1:500-1:20,000)
Alternate names:
Catalog # Size Price (AUD) Your Price (AUD) Qty
A24458 179.00
Check your price
  • F(ab')2-Goat anti-Human IgA Secondary Antibody, AP conjugate for Western Blot, IHC and ELISA
  • Host Goat
  • Target Species Human
  • Conjugate AP
  • Catalog # A24461
  • Size 500µg
Applications
ELISA (1:500-1:20,000), Immunohistochemistry (1:500-1:20,000), Western Blot (1:500-1:20,000)
Alternate names:
Catalog # Size Price (AUD) Your Price (AUD) Qty
A24461 214.00
Check your price
  • Goat anti-Human IgA Cross-Adsorbed Secondary Antibody, Biotin conjugate for IF, Flow and ELISA
  • Host Goat
  • Target Species Human
  • Conjugate Biotin
  • Catalog # A18791
  • Size 1mg
Applications
ELISA (1:500-1:20,000), Flow Cytometry (1:500-1:20,000), Immunofluorescence (1:500-1:20,000)
Alternate names:
Catalog # Size Price (AUD) Your Price (AUD) Qty
A18791 136.00
Check your price
  • Goat anti-Human IgA Cross-Adsorbed Secondary Antibody, TRITC conjugate for IF
  • Host Goat
  • Target Species Human
  • Conjugate TRITC
  • Catalog # A18792
  • Size 1mg
Applications
Immunofluorescence (1:500-1:20,000)
Alternate names:
Catalog # Size Price (AUD) Your Price (AUD) Qty
A18792 118.00
Check your price