1 - 7 of 7 products displayed
  • HIV1 gp41 Polyclonal Antibody, HRP conjugate for Western Blot and ELISA
  • Host Goat
  • Target Species Virus
  • Conjugate HRP
  • Catalog # PA173099
  • Size 1 mL
Applications
ELISA (1:200-1:1,000), Western Blot (Assay Dependent)
Alternate names:
Human immunodeficiency virus, HIV Glycoprotein 41, HIV-1 Glycoprotein 41
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA173099 375.00
Check your price
  • HIV1 gp120 Polyclonal Antibody for Western Blot
  • Host Rabbit
  • Target Species Virus
  • Conjugate Unconjugated
  • Catalog # PA520488
  • Size 100 ug
Applications
Western Blot (0.5-1 ug/ml)
Alternate names:
Human immunodeficiency virus, HIV Glycoprotein 120, HIV-1 Glycoprotein 120
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA520488 349.00
Check your price
  • HIV1 gp41 Monoclonal Antibody for Western Blot
  • Host Mouse
  • Target Species Virus
  • Conjugate Unconjugated
  • Catalog # MA17038
  • Size 1 mL
Applications
Western Blot (1:10-1:50)
Alternate names:
Human immunodeficiency virus, HIV Glycoprotein 41, HIV-1 Glycoprotein 41
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA17038 375.00
Check your price
  • HIV1 gp41 Monoclonal Antibody for Western Blot, IP and ELISA
  • Host Mouse
  • Target Species Virus
  • Conjugate Unconjugated
  • Catalog # MA17590
  • Size 100 ug
Applications
ELISA (Assay Dependent), Immunoprecipitation (Assay Dependent), Western Blot (Assay Dependent)
Alternate names:
Human immunodeficiency virus, HIV Glycoprotein 41, HIV-1 Glycoprotein 41
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA17590 309.00
Check your price
  • HIV1 gp41 Polyclonal Antibody, Biotin conjugate for Western Blot
  • Host Goat
  • Target Species Virus
  • Conjugate Biotin
  • Catalog # PA173100
  • Size 1 mL
Applications
Western Blot (Assay Dependent)
Alternate names:
Human immunodeficiency virus, HIV Glycoprotein 41, HIV-1 Glycoprotein 41
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA173100 375.00
Check your price
  • HIV1 gp41 Polyclonal Antibody for Western Blot
  • Host Goat
  • Target Species Virus
  • Conjugate Unconjugated
  • Catalog # PA17219
  • Size 1 mL
Applications
Western Blot (Assay Dependent)
Alternate names:
Human immunodeficiency virus, HIV Glycoprotein 41, HIV-1 Glycoprotein 41
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA17219 375.00
Check your price
  • HIV1 gp120 Polyclonal Antibody for Western Blot
  • Host Goat
  • Target Species Virus
  • Conjugate Unconjugated
  • Catalog # PA520584
  • Size 100 ug
Applications
Western Blot (1 µg/ml)
Alternate names:
Human immunodeficiency virus, HIV Glycoprotein 120, HIV-1 Glycoprotein 120
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA520584 349.00
Check your price