1 - 30 of 30 products displayed
  • IFNAR2 Monoclonal Antibody for Neu
  • Host Mouse
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # 213851
  • Size 50 ug
Applications
Blocking Assay (Assay Dependent), Neutralization (Assay Dependent)
Alternate names:
Type I interferon receptor 2; Interferon alpha binding protein
Catalog # Size Price (AUD) Your Price (AUD) Qty
213851 475.00
Check your price
  • IFNAR1 Polyclonal Antibody for Western Blot and Flow
  • Host Goat
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # PA124777
  • Size 25 µg
Applications
Flow Cytometry (3-10 µg), Western Blot (0.1-0.2 µg/ml)
Alternate names:
Type I interferon receptor 1; Cytokine receptor family 2 member 1; Cytokine receptor class-II member 1
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA124777 349.00
Check your price
  • Phospho-IFNAR1 (Tyr466) Polyclonal Antibody for IHC (P) and pep-ELISA
  • Host Rabbit
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # PA538502
  • Size 100 µg
Applications
Immunohistochemistry (Paraffin) (1:50-1:100), peptide-ELISA (1:10,000)
Alternate names:
Type I interferon receptor 1; Cytokine receptor family 2 member 1; Cytokine receptor class-II member 1
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA538502 309.00
Check your price
  • IFNAR1 Polyclonal Antibody for IF, ICC and IHC (P)
  • Host Rabbit
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # PA538503
  • Size 100 µg
Applications
Immunocytochemistry (1:100-1:500), Immunofluorescence (1:100-1:500), Immunohistochemistry (Paraffin) (1:50-1:100)
Alternate names:
Type I interferon receptor 1; Cytokine receptor family 2 member 1; Cytokine receptor class-II member 1
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA538503 309.00
Check your price
  • IFNAR1 Monoclonal Antibody, PE conjugate for Flow
  • Host Mouse
  • Target Species Human
  • Conjugate PE
  • Catalog # MA523630
  • Size 100 Tests
Applications
Flow Cytometry (10 µl/10^6 cells)
Alternate names:
Type I interferon receptor 1; Cytokine receptor family 2 member 1; Cytokine receptor class-II member 1
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA523630 309.00
Check your price
  • IFNAR2 Polyclonal Antibody for Western Blot, IF and ICC
  • Host Rabbit
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # PA528303
  • Size 100 µL
Applications
Immunocytochemistry (1:100-1:1000), Immunofluorescence (1:100-1:1000), Western Blot (1:500-1:3000)
Alternate names:
Type I interferon receptor 2; Interferon alpha binding protein
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA528303 349.00
Check your price
  • IFNAR2 Polyclonal Antibody for Western Blot
  • Host Chicken
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # PA19544
  • Size 500 ug
Applications
Western Blot (1:500-1:5,000)
Alternate names:
Type I interferon receptor 2; Interferon alpha binding protein
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA19544 225.00
Check your price
  • IFNAR1 Polyclonal Antibody for Western Blot
  • Host Rabbit
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # PA521462
  • Size 100 µL
Applications
Western Blot (1:500-1:3000)
Alternate names:
Type I interferon receptor 1; Cytokine receptor family 2 member 1; Cytokine receptor class-II member 1
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA521462 349.00
Check your price
  • Phospho-IFNAR1 (Ser535, Ser539) Polyclonal Antibody for Western Blot
  • Host Rabbit
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # PA537789
  • Size 100 µl
Applications
Western Blot (1:500-1:1000)
Alternate names:
Type I interferon receptor 1; Cytokine receptor family 2 member 1; Cytokine receptor class-II member 1
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA537789 309.00
Check your price
  • IFN alpha Polyclonal Antibody for Neu
  • Host Sheep
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # 311001
  • Size 1x10^5 units
Applications
Neutralization (Assay Dependent)
Alternate names:
Interferon alpha-D
Catalog # Size Price (AUD) Your Price (AUD) Qty
311001 269.00
Check your price
  • IFN alpha Polyclonal Antibody for Neu
  • Host Rabbit
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # 311301
  • Size 2x10^4 units
Applications
Neutralization (Assay Dependent)
Alternate names:
Interferon alpha-D
Catalog # Size Price (AUD) Your Price (AUD) Qty
311301 269.00
Check your price
  • Phospho-IFNAR1 (Ser535, Ser539) Polyclonal Antibody for Western Blot and IHC
  • Host Rabbit
  • Target Species Human, Rat
  • Conjugate Unconjugated
  • Catalog # PA14627
  • Size 100 µL
Applications
Immunohistochemistry (1:1000), Western Blot (1:1000)
Alternate names:
Type I interferon receptor 1; Cytokine receptor family 2 member 1; Cytokine receptor class-II member 1
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA14627 349.00
Check your price
  • IFN beta Polyclonal Antibody for Western Blot, IHC, IP, ELISA and Neu
  • Host Sheep
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # AHC4024
  • Size 2x10^4 NU
Applications
ELISA (Assay Dependent), Immunohistochemistry (Assay Dependent), Immunoprecipitation (Assay Dependent), Neutralization (Assay Dependent), Western Blot (Assay Dependent)
Alternate names:
Fibroblast interferon
Catalog # Size Price (AUD) Your Price (AUD) Qty
AHC4024 309.00
Check your price
  • LXR alpha/beta Polyclonal Antibody for Western Blot
  • Host Rabbit
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # PA1331B
  • Size 200 µg
Applications
Western Blot (1:500)
Alternate names:
Nuclear receptor subfamily 1 group H member 3; Liver X receptor alpha; Ubiquitously-expressed nuclear receptor; Nuclear receptor subfamily 1 group H member 2; Nuclear receptor NER; Liver X receptor beta
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA1331B 349.00
Check your price
  • IFN alpha Monoclonal Antibody for FN
  • Host Mouse
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # MA146034
  • Size 100 µg
Applications
Functional Assay (Assay-Dependent)
Alternate names:
Interferon alpha-D
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA146034 309.00
Check your price
  • IFN beta Monoclonal Antibody for ELISA and Neu
  • Host Mouse
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # 214001
  • Size 50 ug
Applications
ELISA (Assay Dependent), Neutralization (Assay Dependent)
Alternate names:
Fibroblast interferon
Catalog # Size Price (AUD) Your Price (AUD) Qty
214001 275.00
Check your price
  • IFN beta Polyclonal Antibody for Neu
  • Host Rabbit
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # 314101
  • Size 2x10^4 units
Applications
Neutralization (Assay Dependent)
Alternate names:
Fibroblast interferon
Catalog # Size Price (AUD) Your Price (AUD) Qty
314101 275.00
Check your price
  • IFN beta Monoclonal Antibody for ELISA
  • Host Mouse
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # 214051
  • Size 50 ug
Applications
ELISA (Assay Dependent)
Alternate names:
Fibroblast interferon
Catalog # Size Price (AUD) Your Price (AUD) Qty
214051 275.00
Check your price
  • IFNAR2 Polyclonal Antibody for Western Blot and Neu
  • Host Goat
  • Target Species Mouse
  • Conjugate Unconjugated
  • Catalog # PA547062
  • Size 100 µg
Applications
Neutralization (2-10 µg/ml), Western Blot (0.1 µg/ml)
Alternate names:
Type I interferon receptor 2
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA547062 349.00
Check your price
  • CaMKII alpha/beta/delta Polyclonal Antibody for IHC (P)
  • Host Rabbit
  • Target Species Rat, Human, Mouse
  • Conjugate Unconjugated
  • Catalog # PA538239
  • Size 100 µg
Applications
Immunohistochemistry (Paraffin) (1:50-1:100)
Alternate names:
Calcium/calmodulin-dependent protein kinase type II subunit alpha, CaM kinase II subunit alpha, CaMK-II subunit alpha, CAMK2A, CAMKA, KIAA0968, CaMK2-a, CaMK2a, CaMK-2a, Calcium/calmodulin-dependent protein kinase type II subunit delta, CaM kinase II subunit delta, CaMK-II subunit delta, CAMK2D, CAMKD, CaMK2- delta, CaMK2 delta, CaMK-2 delta
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA538239 309.00
Check your price
  • Phospho-CaMKII alpha/beta/delta (Thr305) Polyclonal Antibody for Western Blot and IHC (P)
  • Host Rabbit
  • Target Species Human, Mouse
  • Conjugate Unconjugated
  • Catalog # PA537832
  • Size 100 µl
Applications
Immunohistochemistry (Paraffin) (1:50-1:100), Western Blot (1:500-1:1000)
Alternate names:
calcium/calmodulin-dependent protein kinase (CaM kinase) II alpha, calcium/calmodulin-dependent protein kinase II alpha, calcium/calmodulin-dependent protein kinase II alpha-B subunit, calcium/calmodulin-dependent protein kinase type II alpha chain, Calci, calcium/calmodulin-dependent protein kinase (CaM kinase) II delta, calcium/calmodulin-dependent protein kinase II delta, calcium/calmodulin-dependent protein kinase type II delta chain, Calcium/calmodulin-dependent protein kinase type II subunit delta, Ca, CAMKD, CaM kinase II delta subunit, CaM-kinase II delta chain, CaMK-II delta subunit, caM kinase II subunit delta, caMK-II subunit delta, calcium/calmodulin-dependent protein kinase (CaM kinase) II delta, calcium/calmodulin-dependent protein kinase type II delta chain, calcium/calmodulin-dependent protein kinase type II subunit delta
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA537832 309.00
Check your price
  • IFN beta Polyclonal Antibody for Western Blot, IF, ICC and IHC
  • Host Rabbit
  • Target Species Human, Mouse
  • Conjugate Unconjugated
  • Catalog # PA520390
  • Size 100 µg
Applications
Immunocytochemistry (20 ug/ml), Immunofluorescence (20 ug/ml), Immunohistochemistry (5 ug/ml), Western Blot (5 ug/ml)
Alternate names:
Fibroblast interferon
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA520390 349.00
Check your price
  • IFN alpha Monoclonal Antibody for Western Blot, IHC (F), ELISA and Neu
  • Host Mouse
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # MA135482
  • Size 100 µg
Applications
Blocking Assay (Assay dependent), ELISA (Assay dependent.), Immunohistochemistry (Frozen) (Assay dependent), Western Blot (Assay dependent.)
Alternate names:
Interferon alpha-D
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA135482 309.00
Check your price
  • LXR alpha Polyclonal Antibody for Western Blot, IF, ICC and Flow
  • Host Rabbit
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # PA1330
  • Size 200 µg
Applications
Immunocytochemistry (1:50-1:500), Immunofluorescence (1:50-1:500), Western Blot (1:300)
Alternate names:
Nuclear receptor subfamily 1 group H member 3; Liver X receptor alpha
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA1330 375.00
Check your price
  • TNF alpha Monoclonal Antibody for Western Blot, IF, ICC, ELISA and Neu
  • Host Mouse
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # MA523720
  • Size 500 µg
Applications
ELISA (0.1-0.4 µg/ml (Detection), 2-8 µg/ml (Capture)), Immunocytochemistry (8-25 µg/ml), Immunofluorescence (8-25 µg/ml), Neutralization (0.04-0.08 µg/ml), Western Blot (1 µg/ml)
Alternate names:
Tumor necrosis factor ligand superfamily member 2; TNF-alpha; Cachectin
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA523720 375.00
Check your price
  • LXR alpha Monoclonal Antibody for Western Blot and ELISA
  • Host Mouse
  • Target Species Human, Rat
  • Conjugate Unconjugated
  • Catalog # 418300
  • Size 100 µg
Applications
ELISA (Assay Dependent), Western Blot (Assay Dependent)
Alternate names:
Nuclear receptor subfamily 1 group H member 3; Liver X receptor alpha
Catalog # Size Price (AUD) Your Price (AUD) Qty
418300 426.00
Check your price
  • IL-1 beta Monoclonal Antibody, FITC conjugate for Flow
  • Host Mouse
  • Target Species Human
  • Conjugate FITC
  • Catalog # MA523657
  • Size 100 Tests
Applications
Flow Cytometry (10 µl/10^6 cells)
Alternate names:
Catabolin
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA523657 349.00
Check your price
  • IL-1 beta Monoclonal Antibody, PE conjugate for Flow
  • Host Mouse
  • Target Species Human
  • Conjugate PE
  • Catalog # MA523546
  • Size 100 Tests
Applications
Flow Cytometry (10 µl/10^6 cells)
Alternate names:
Catabolin
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA523546 239.00
Check your price
  • IL-1 beta Monoclonal Antibody for Western Blot, IF, ICC, ELISA and Neu
  • Host Mouse
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # MA523691
  • Size 500 µg
Applications
ELISA (0.1-0.4 µg/ml (Detection), 2-8 µg/ml (Capture)), Immunocytochemistry (8-25 µg/ml), Immunofluorescence (8-25 µg/ml), Neutralization (0.05-0.2 µg/ml), Western Blot (1 µg/ml)
Alternate names:
Catabolin
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA523691 375.00
Check your price
  • IL-1 beta Monoclonal Antibody, APC conjugate for Flow
  • Host Mouse
  • Target Species Human
  • Conjugate APC
  • Catalog # MA523597
  • Size 100 Tests
Applications
Flow Cytometry (Assay-Dependent)
Alternate names:
Catabolin
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA523597 289.00
Check your price