1 - 8 of 8 products displayed
  • lys-Plasminogen (human) for Ctrl
  • Host Human
  • Target Species Not Applicable
  • Conjugate Unconjugated
  • Catalog # RP43081
  • Size 1mg
Applications
Control (Assay Dependent)
Alternate names:
Catalog # Size Price (AUD) Your Price (AUD) Qty
RP43081 249.00
Check your price
  • Urokinase Polyclonal Antibody for Western Blot, IHC (F), IHC (P) and ELISA
  • Host Goat
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # PA136015
  • Size 500µg
Applications
ELISA (1:5000), Immunohistochemistry (Paraffin, Frozen) (1:50), Western Blot (Assay dependent.)
Alternate names:
ATF, UPA, URK, u-PA
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA136015 449.00
Check your price
  • Urokinase Polyclonal Antibody for Western Blot and IHC (P)
  • Host Rabbit
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # PA534790
  • Size 100µL
Applications
Immunohistochemistry (Paraffin) (1:100-1:1000), Western Blot (1:500-1:3000)
Alternate names:
ATF, BDPLT5, QPD, UPA, URK, u-PA, U-plasminogen activator, plasminogen activator, urinary, urokinase-type plasminogen activator
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA534790 349.00
Check your price
  • Urokinase Polyclonal Antibody for Western Blot and IHC (P)
  • Host Rabbit
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # PA534638
  • Size 100µL
Applications
Immunohistochemistry (Paraffin) (1:100-1:1000), Western Blot (1:500-1:3000)
Alternate names:
ATF, BDPLT5, QPD, UPA, URK, u-PA, U-plasminogen activator, plasminogen activator, urinary, urokinase-type plasminogen activator
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA534638 349.00
Check your price
  • Urokinase Polyclonal Antibody for Western Blot, IHC (F), IHC (P), ELISA and RIA
  • Host Rabbit
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # PA136166
  • Size 500µg
Applications
ELISA (1:5000), Immunohistochemistry (Paraffin, Frozen) (1:50), Radioimmune Assays (Assay dependent), Western Blot (Assay dependent.)
Alternate names:
ATF, UPA, URK, u-PA
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA136166 375.00
Check your price
  • Plasma Kallikrein Polyclonal Antibody for Western Blot
  • Host Rabbit
  • Target Species Dog, Horse, Human, Pig, Rat
  • Conjugate Unconjugated
  • Catalog # PA542752
  • Size 100µl
Applications
Western Blot (2.5 µg/ml)
Alternate names:
Plasma prekallikrein; Kininogenin; Fletcher factor
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA542752 309.00
Check your price
  • Plasma Kallikrein Polyclonal Antibody for Western Blot
  • Host Rabbit
  • Target Species Human, Mouse
  • Conjugate Unconjugated
  • Catalog # PA534876
  • Size 100µL
Applications
Western Blot (1:5000-1:20,000)
Alternate names:
Plasma prekallikrein; Kininogenin; Fletcher factor
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA534876 349.00
Check your price
  • Plasma Kallikrein Polyclonal Antibody for Western Blot and IP
  • Host Goat
  • Target Species Mouse
  • Conjugate Unconjugated
  • Catalog # PA547334
  • Size 100µg
Applications
Immunoprecipitation (25 µg/ml), Western Blot (0.1 µg/ml)
Alternate names:
Plasma prekallikrein; Kininogenin; Fletcher factor
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA547334 349.00
Check your price