1 - 2 of 2 products displayed
  • MARCO Polyclonal Antibody for Western Blot, IHC and Flow
  • Host Rabbit
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # PA526888
  • Size 400 µL
Applications
Flow Cytometry (1:10-1:50), Immunohistochemistry (1:50-1:100), Western Blot (1:1000)
Alternate names:
Scavenger receptor class A member 2; Macrophage receptor with collagenous structure
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA526888 349.00
Check your price
  • MARCO Monoclonal Antibody for Western Blot, IF, IHC (F), Flow and IP
  • Host Mouse
  • Target Species Bovine, Human
  • Conjugate Unconjugated
  • Catalog # MA140315
  • Size 100 µg
Applications
Flow Cytometry (Assay Dependent), Immunofluorescence (Assay Dependent), Immunohistochemistry (Frozen) (Assay Dependent), Immunoprecipitation (Assay Dependent), Western Blot (Assay Dependent)
Alternate names:
Scavenger receptor class A member 2; Macrophage receptor with collagenous structure
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA140315 600.00
Check your price