1 - 30 of 30 products displayed
  • Ki-67 Antibody (Monoclonal, 12H15 L5)
  • Host Rabbit
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # 701198
  • Size 100 µg
Applications
ChIP assay (1 ul), Flow Cytometry (1:100-1:500), Immunocytochemistry (1:100-1:500), Immunofluorescence (1:100-1:500)
Alternate names:
antigen KI-67, proliferation-related Ki-67 antigen, RP11-380J17.2, KIA
Catalog # Size Price (AUD) Your Price (AUD) Qty
701198 303.00
Check your price
  • CD171 Antibody (Monoclonal, UJ127)
  • Host Mouse
  • Target Species Human, Mouse
  • Conjugate Unconjugated
  • Catalog # MA514140
  • Size 500 µL
Applications
Immunohistochemistry (1:1000), Immunohistochemistry (Paraffin) (1:100), Immunoprecipitation (2 µg/mg protein lysate), Western Blot (1-2 µg/ml)
Alternate names:
CAML1, CD171, HSAS, HSAS1, MASA, MIC5, N-CAML1, S10, SPG1
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA514140 349.00
Check your price
  • Ki-67 Antibody (Monoclonal, 8D5)
  • Host Mouse
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # MA515690
  • Size 100 µL
Applications
Immunohistochemistry (1/200-1/1000), Western Blot (1/500-1/2000)
Alternate names:
KIA; Ki-67; MKI67
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA515690 349.00
Check your price
  • Ki-67 Antibody (Monoclonal, 4A1)
  • Host Mouse
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # MA515525
  • Size 100 µL
Applications
Immunohistochemistry (1/200-1/1000)
Alternate names:
KIA; Ki-67; MKI67
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA515525 349.00
Check your price
  • CD99 Antibody (Monoclonal, HO36-1.1)
  • Host Mouse
  • Target Species Human, Rat
  • Conjugate Unconjugated
  • Catalog # MA512954
  • Size 500 µL
Applications
Flow Cytometry (2 ug/test), Immunocytochemistry (1:10-1:100), Immunofluorescence (1:10-1:100), Immunohistochemistry (Paraffin) (1:100-200), Western Blot (1:100-1:500)
Alternate names:
T-cell surface glycoprotein E2; Protein MIC2; E2 antigen; 120000000
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA512954 309.00
Check your price
  • CD99 Antibody (Monoclonal, O13)
  • Host Mouse
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # MA512287
  • Size 500 µL
Applications
Immunohistochemistry (Paraffin) (1-2 µg/ml)
Alternate names:
T-cell surface glycoprotein E2; Protein MIC2; E2 antigen; 120000000
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA512287 309.00
Check your price
  • CD56 Antibody, PE (Monoclonal, MEM-188)
  • Host Mouse
  • Target Species Human, Non-human primate
  • Conjugate PE
  • Catalog # MA119638
  • Size 100 tests
Applications
Flow Cytometry (20ul/100ul whole blood)
Alternate names:
NCAM1, NCAM, CD-56, MSK39, neural cell adhesion molecule 1
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA119638 275.00
Check your price
  • CD56 Antibody (Monoclonal, MEM-188)
  • Host Mouse
  • Target Species Human, Non-human primate
  • Conjugate Unconjugated
  • Catalog # MA119129
  • Size 100 µg
Applications
Flow Cytometry (3µg/ml), Immunohistochemistry (Paraffin) (1:50), Immunoprecipitation (Assay Dependent)
Alternate names:
NCAM1, NCAM, CD-56, MSK39, neural cell adhesion molecule 1
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA119129 199.00
Check your price
  • CD171 Antibody, Biotin (Monoclonal, UJ127)
  • Host Mouse
  • Target Species Human
  • Conjugate Biotin
  • Catalog # MA514137
  • Size 500 µL
Applications
Immunohistochemistry (Paraffin) (1:100), Immunoprecipitation (2 µg/ml), Western Blot (1-2 µg/ml)
Alternate names:
CAML1, CD171, HSAS, HSAS1, MASA, MIC5, N-CAML1, S10, SPG1
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA514137 399.00
Check your price
  • Ki-67 Antibody (Monoclonal, SP6)
  • Host Rabbit
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # MA514520
  • Size 500 µL
Applications
Flow Cytometry (3-5 µg x 10^6 cells), Immunohistochemistry (Paraffin) (1:100-200)
Alternate names:
antigen KI-67, proliferation-related Ki-67 antigen
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA514520 375.00
Check your price
  • Aggrecan Antibody (Monoclonal, BC-3)
  • Host Mouse
  • Target Species Bovine, Dog, Horse, Cat, Guinea pig, Human, Sheep, Pig, Rabbit, Rat
  • Conjugate Unconjugated
  • Catalog # MA316888
  • Size 100 µg
Applications
ELISA (1:1000-1:5000), Immunofluorescence (1:100-1:500), Immunohistochemistry (Frozen) (1:10-1:2000), Immunohistochemistry (Paraffin) (1:10-1:2000), Western Blot (1:500-1:1000)
Alternate names:
Cartilage-specific proteoglycan core protein
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA316888 309.00
Check your price
  • CD171 Antibody (Monoclonal, L1.1)
  • Host Mouse
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # 375100
  • Size 100 µg
Applications
ELISA (Assay Dependent), Immunocytochemistry (Assay Dependent), Immunofluorescence (Assay Dependent), Immunoprecipitation (Assay Dependent)
Alternate names:
CAML1, CD171, HSAS, HSAS1, MASA, MIC5, N-CAML1, S10, SPG1
Catalog # Size Price (AUD) Your Price (AUD) Qty
375100 309.00
Check your price
  • CD59 Antibody, PE (Monoclonal, MEM-43)
  • Host Mouse
  • Target Species Human
  • Conjugate PE See Additional Formats FITC
  • Catalog # MHCD59044
  • Size 2 ml
Applications
Flow Cytometry (Assay-Dependent)
Alternate names:
Protectin; Membrane inhibitor of reactive lysis; Membrane attack complex inhibition factor; MEM43 antigen; MAC-inhibitory protein; 20 kDa homologous restriction factor; 1F5 antigen
Catalog # Size Price (AUD) Your Price (AUD) Qty
MHCD59044 868.00
Check your price
  • CD59 Antibody, PE (Monoclonal, MEM-43)
  • Host Mouse
  • Target Species Human
  • Conjugate PE See Additional Formats FITC
  • Catalog # MHCD5904
  • Size 0.5 ml
Applications
Flow Cytometry (Assay-Dependent)
Alternate names:
Protectin; Membrane inhibitor of reactive lysis; Membrane attack complex inhibition factor; MEM43 antigen; MAC-inhibitory protein; 20 kDa homologous restriction factor; 1F5 antigen
Catalog # Size Price (AUD) Your Price (AUD) Qty
MHCD5904 258.00
Check your price
  • CD56 Antibody, PE (Monoclonal, MEM-188)
  • Host Mouse
  • Target Species Human, Non-human primate
  • Conjugate PE
  • Catalog # MA121343
  • Size 100 tests
Applications
Flow Cytometry (Assay dependent)
Alternate names:
NCAM1, NCAM, CD-56, MSK39, neural cell adhesion molecule 1
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA121343 425.00
Check your price
  • CD171 Antibody (Monoclonal, UJ127.11)
  • Host Mouse
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # MA146044
  • Size 100 µg
Applications
Immunohistochemistry (Paraffin) (1:10-1:500), Immunoprecipitation (1:10-1:500), Western Blot (1:100-1:2000)
Alternate names:
CAML1, CD171, HSAS, HSAS1, MASA, MIC5, N-CAML1, S10, SPG1
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA146044 375.00
Check your price
  • CD56 Antibody, PE-Cyanine5.5 (Monoclonal, MEM-188)
  • Host Mouse
  • Target Species Human
  • Conjugate PE-Cyanine5.5 See Additional Formats Unconjugated Biotin FITC PE Alexa Fluor 488 Alexa Fluor 647 TRI-COLOR PE-Texas Red
  • Catalog # MHCD5618
  • Size 0.5 ml
Applications
Flow Cytometry (Assay-Dependent)
Alternate names:
NCAM1, NCAM, CD-56, MSK39, neural cell adhesion molecule 1
Catalog # Size Price (AUD) Your Price (AUD) Qty
MHCD5618 317.00
Check your price
  • CD56 Antibody (Monoclonal, MEM-188)
  • Host Mouse
  • Target Species Human
  • Conjugate Unconjugated See Additional Formats Biotin FITC PE Alexa Fluor 488 Alexa Fluor 647 TRI-COLOR PE-Cyanine5.5 PE-Texas Red
  • Catalog # MHCD5600
  • Size 0.5 ml
Applications
Flow Cytometry (Assay-Dependent)
Alternate names:
NCAM1, NCAM, CD-56, MSK39, neural cell adhesion molecule 1
Catalog # Size Price (AUD) Your Price (AUD) Qty
MHCD5600 158.00
Check your price
  • CD200 Antibody, APC (Monoclonal, 325516)
  • Host Mouse
  • Target Species Human
  • Conjugate APC
  • Catalog # MA523575
  • Size 100 Tests
Applications
Flow Cytometry (10 µl/10^6 cells)
Alternate names:
My033, MOX1, MOX2, MRC, OX-2, CD200 antigen, MRC OX-2 antigen, OX-2 membrane glycoprotein, antigen identified by monoclonal antibody MRC OX-2
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA523575 249.00
Check your price
  • CD200 Antibody, PE (Monoclonal, 325516)
  • Host Mouse
  • Target Species Human
  • Conjugate PE
  • Catalog # MA523576
  • Size 100 Tests
Applications
Flow Cytometry (10 µl/10^6 cells)
Alternate names:
My033, MOX1, MOX2, MRC, OX-2, CD200 antigen, MRC OX-2 antigen, OX-2 membrane glycoprotein, antigen identified by monoclonal antibody MRC OX-2
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA523576 249.00
Check your price
  • CD200 Antibody (Monoclonal, 325516)
  • Host Mouse
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # MA524048
  • Size 100 µg
Applications
Flow Cytometry (0.25 µg/10^6 cells)
Alternate names:
My033, MOX1, MOX2, MRC, OX-2, CD200 antigen, MRC OX-2 antigen, OX-2 membrane glycoprotein, antigen identified by monoclonal antibody MRC OX-2
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA524048 275.00
Check your price
  • CD59 Antibody (Monoclonal, 8D2B8)
  • Host Mouse
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # MA517046
  • Size 100 ug
Applications
Flow Cytometry (1:200-1:400), Immunocytochemistry (1:50 -1:200), Immunofluorescence (1:50-200), Immunohistochemistry (1:200-1:1000), Western Blot (1:500-1:2000)
Alternate names:
Protectin; Membrane inhibitor of reactive lysis; Membrane attack complex inhibition factor; MEM43 antigen; MAC-inhibitory protein; 20 kDa homologous restriction factor; 1F5 antigen
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA517046 309.00
Check your price
  • CD200 Antibody, FITC (Monoclonal, OX-104)
  • Host Mouse
  • Target Species Human
  • Conjugate FITC
  • Catalog # MA516897
  • Size 100 ug
Applications
Flow Cytometry (Neat-1:5)
Alternate names:
My033, MOX1, MOX2, MRC, OX-2, CD200 antigen, MRC OX-2 antigen, OX-2 membrane glycoprotein, antigen identified by monoclonal antibody MRC OX-2
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA516897 475.00
Check your price
  • CD99 Antibody (Monoclonal, SP119)
  • Host Rabbit
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # MA516416
  • Size 500 µL
Applications
Immunohistochemistry (Paraffin) (1:200)
Alternate names:
T-cell surface glycoprotein E2; Protein MIC2; E2 antigen; 120000000
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA516416 499.00
Check your price
  • CD56 Antibody (Monoclonal, 3H15L12)
  • Host Rabbit
  • Target Species Human, Mouse, Rat
  • Conjugate Unconjugated
  • Catalog # 701379
  • Size 100 µg
Applications
Flow Cytometry (1 ug/ml X 10^6 cells), Immunocytochemistry (1 ug/ml), Immunofluorescence (1 ug/ml), Western Blot (0.5-2 ug/ml)
Alternate names:
NCAM1, NCAM, CD-56, MSK39, neural cell adhesion molecule 1
Catalog # Size Price (AUD) Your Price (AUD) Qty
701379 303.00
Check your price
  • CD46 Antibody, APC (Monoclonal, MEM-258)
  • Host Mouse
  • Target Species Bovine, Human
  • Conjugate APC
  • Catalog # MA119784
  • Size 100 tests
Applications
Flow Cytometry (Assay Dependent)
Alternate names:
Trophoblast leukocyte common antigen; TLX
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA119784 309.00
Check your price
  • JAK3 Antibody (Monoclonal, 5H2)
  • Host Mouse
  • Target Species Human, Mouse
  • Conjugate Unconjugated
  • Catalog # MA515561
  • Size 100 µL
Applications
Flow Cytometry (1/200-1/400), Immunocytochemistry (1:200-1:1000), Immunofluorescence (1/200-1/1000), Western Blot (1/500-1/2000)
Alternate names:
Janus kinase 3; Leukocyte janus kinase
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA515561 349.00
Check your price
  • CD200 Antibody (Monoclonal, 325531)
  • Host Mouse
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # MA523912
  • Size 100 µg
Applications
Blocking Assay (Assay-Dependent), ELISA (0.5-2 µg/ml (Detection), 2-8 µg/ml (Capture))
Alternate names:
My033, MOX1, MOX2, MRC, OX-2, CD200 antigen, MRC OX-2 antigen, OX-2 membrane glycoprotein, antigen identified by monoclonal antibody MRC OX-2
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA523912 275.00
Check your price
  • Galectin 10 Antibody (Monoclonal, 561603)
  • Host Mouse
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # MA524251
  • Size 100 µg
Applications
Flow Cytometry (2.5 µg/10^6 cells), Western Blot (1 µg/ml)
Alternate names:
Lysolecithin acylhydrolase; Eosinophil lysophospholipase; Charcot-Leyden crystal protein
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA524251 275.00
Check your price
  • CD56 Antibody, Biotin (Monoclonal, MEM-188)
  • Host Mouse
  • Target Species Human, Non-human primate
  • Conjugate Biotin
  • Catalog # MA119502
  • Size 100 µg
Applications
Flow Cytometry (5 µg/ml)
Alternate names:
NCAM1, NCAM, CD-56, MSK39, neural cell adhesion molecule 1
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA119502 249.00
Check your price