1 - 30 of 30 products displayed
  • Prolactin Receptor Monoclonal Antibody for IHC (P)
  • Host Mouse
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # MA511955
  • Size 500µL
Applications
Immunohistochemistry (Paraffin) (1-2 µg/ml)
Alternate names:
PRL-R, delta 4-SF1b truncated prolactin receptor, delta 4-delta 7/11 truncated prolactin receptor, hPRL receptor, prolactin receptor delta 7/11, secreted prolactin binding protein
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA511955 349.00
Check your price
  • Prolactin Receptor Monoclonal Antibody for Western Blot, IF, ICC, IHC, Flow and IP
  • Host Mouse
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # 359200
  • Size 100µg
Applications
Immunohistochemistry (Assay Dependent), Immunoprecipitation (Assay Dependent), Western Blot (Assay Dependent)
Alternate names:
hPRLrI, PRL-R, hPRL receptor, secreted prolactin binding protein
Catalog # Size Price (AUD) Your Price (AUD) Qty
359200 309.00
Check your price
  • Growth Hormone Monoclonal Antibody for Western Blot and IHC (P)
  • Host Mouse
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # MA511926
  • Size 500µL
Applications
Immunohistochemistry (Paraffin) (1:2000-1:4000), Western Blot (1-2 µg/ml)
Alternate names:
Pituitary growth hormone; Growth hormone 1; Growth hormone
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA511926 309.00
Check your price
  • STAT5 alpha/beta Polyclonal Antibody for Western Blot
  • Host Rabbit
  • Target Species Hamster, Human, Mouse
  • Conjugate Unconjugated
  • Catalog # OPA103010
  • Size 100µg
Applications
Western Blot (Assay-Dependent)
Alternate names:
Mammary gland factor
Catalog # Size Price (AUD) Your Price (AUD) Qty
OPA103010 349.00
Check your price
  • Annexin A1 Polyclonal Antibody for Western Blot, IF, ICC, IHC (P), Flow and ELISA
  • Host Rabbit
  • Target Species Human, Mouse, Rat
  • Conjugate Unconjugated
  • Catalog # 713400
  • Size 100µL
Applications
ELISA (Assay Dependent), Flow Cytometry (3-5 µg/million cells), Immunocytochemistry (1µg/mL), Immunofluorescence (1µg/mL), Immunohistochemistry (Paraffin) (1:20-1:200), Western Blot (1:500-1:2000)
Alternate names:
p35; Phospholipase A2 inhibitory protein; Lipocortin I; Chromobindin-9; Calpactin-2; Calpactin II; Annexin-1; Annexin I
Catalog # Size Price (AUD) Your Price (AUD) Qty
713400 307.00
Check your price
  • Phospho-STAT5 alpha (Tyr694) Polyclonal Antibody for Western Blot, IF, ICC, IHC (P), Flow and ChIP
  • Host Rabbit
  • Target Species Human, Mouse
  • Conjugate Unconjugated
  • Catalog # 44390G
  • Size 10blots
Applications
ChIP assay (1-5 µg), Flow Cytometry (3-5 µg/million cells), Immunocytochemistry (1-2 µg/ml), Immunofluorescence (1-2 µg/ml), Immunohistochemistry (Paraffin) (1:10-1:100), Western Blot (Assay Dependent)
Alternate names:
Mammary gland factor
Catalog # Size Price (AUD) Your Price (AUD) Qty
44390G 309.00
Check your price
  • Phospho-STAT5 alpha (Tyr694) Polyclonal Antibody for Western Blot, IHC and ELISA
  • Host Rabbit
  • Target Species Bovine, Human, Mouse, Sheep, Rat
  • Conjugate Unconjugated
  • Catalog # 716900
  • Size 50µg
Applications
ELISA (Assay Dependent), Immunohistochemistry (Assay Dependent), Western Blot (Assay Dependent)
Alternate names:
Mammary gland factor
Catalog # Size Price (AUD) Your Price (AUD) Qty
716900 305.00
Check your price
  • Phospho-STAT5 beta (Tyr694) Monoclonal Antibody for Western Blot and ELISA
  • Host Mouse
  • Target Species Human, Mouse, Sheep, Rat, Bovine
  • Conjugate Unconjugated
  • Catalog # 336000
  • Size 50µg
Applications
ELISA (Assay Dependent), Western Blot (Assay Dependent)
Alternate names:
Signal transducer and activator of transcription 5B, STA5B, STAT5, STAT5B, transcription factor STAT5B
Catalog # Size Price (AUD) Your Price (AUD) Qty
336000 306.00
Check your price
  • PREB Polyclonal Antibody for Western Blot
  • Host Rabbit
  • Target Species Mouse
  • Conjugate Unconjugated
  • Catalog # PA1610
  • Size 100µL
Applications
Western Blot (1:500)
Alternate names:
Mammalian guanine nucleotide exchange factor mSec12
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA1610 349.00
Check your price
  • PITX2 Polyclonal Antibody for Western Blot, IF and ICC
  • Host Rabbit
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # PA511479
  • Size 400µL
Applications
Immunocytochemistry (1:10-1:50), Immunofluorescence (1:10-1:50), Western Blot (1:1000)
Alternate names:
Solurshin; RIEG bicoid-related homeobox transcription factor; Paired-like homeodomain transcription factor 2; Homeobox protein PITX2; ALL1-responsive protein ARP1
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA511479 349.00
Check your price
  • STAT5 alpha Oligoclonal Antibody for Western Blot and IHC (P)
  • Host Rabbit
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # 710073
  • Size 100µg
Applications
Immunohistochemistry (Paraffin) (1:10-1:50), Western Blot (0.5-1µg/ml)
Alternate names:
MGF, STAT5
Catalog # Size Price (AUD) Your Price (AUD) Qty
710073 309.00
Check your price
  • Annexin V Polyclonal Antibody for Western Blot and IHC
  • Host Rabbit
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # PA513523
  • Size 400µL
Applications
Immunohistochemistry (1:10-1:50), Western Blot (1:1000)
Alternate names:
Vascular anticoagulant-alpha; Thromboplastin inhibitor; Placental anticoagulant protein I; Placental anticoagulant protein 4; Lipocortin V; Endonexin II; Calphobindin I; Annexin-5; Annexin V; Anchorin CII
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA513523 349.00
Check your price
  • Annexin A1 Polyclonal Antibody for IHC (P)
  • Host Rabbit
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # PA137083
  • Size 1mL
Applications
Immunohistochemistry (Paraffin) (1:25)
Alternate names:
p35; Phospholipase A2 inhibitory protein; Lipocortin I; Chromobindin-9; Calpactin-2; Calpactin II; Annexin-1; Annexin I
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA137083 449.00
Check your price
  • Annexin A1 Polyclonal Antibody for IHC (P)
  • Host Rabbit
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # PA532264
  • Size 500µL
Applications
Immunohistochemistry (Paraffin) (1:25)
Alternate names:
p35; Phospholipase A2 inhibitory protein; Lipocortin I; Chromobindin-9; Calpactin-2; Calpactin II; Annexin-1; Annexin I
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA532264 349.00
Check your price
  • PITX1 Polyclonal Antibody for Western Blot, IHC (P) and Flow
  • Host Rabbit
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # PA514219
  • Size 400µL
Applications
Flow Cytometry (1:10-1:50), Immunohistochemistry (1:50-1:100), Western Blot (1:1000)
Alternate names:
Paired-like homeodomain transcription factor 1; Homeobox protein PITX1; Hindlimb-expressed homeobox protein backfoot
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA514219 349.00
Check your price
  • ABAT Polyclonal Antibody for Western Blot, IF, ICC and IHC (P)
  • Host Rabbit
  • Target Species Human, Mouse, Rat
  • Conjugate Unconjugated
  • Catalog # PA527198
  • Size 100µL
Applications
Immunocytochemistry (1:100-1:1000), Immunofluorescence (1:100-1:1000), Immunohistochemistry (Paraffin) (1:100-1:1000), Western Blot (1:1000-1:10,000)
Alternate names:
L-AIBAT; Gamma-amino-N-butyrate transaminase; GABA aminotransferase; (S)-3-amino-2-methylpropionate transaminase
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA527198 349.00
Check your price
  • ABAT Polyclonal Antibody for Western Blot
  • Host Rabbit
  • Target Species Bovine, Dog, Horse, Guinea pig, Goat, Human, Mouse, Rabbit, Rat, Yeast, Zebrafish
  • Conjugate Unconjugated
  • Catalog # PA543723
  • Size 100µl
Applications
Western Blot (0.2-1 µg/ml)
Alternate names:
L-AIBAT; Gamma-amino-N-butyrate transaminase; GABA aminotransferase; (S)-3-amino-2-methylpropionate transaminase
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA543723 309.00
Check your price
  • ABAT Polyclonal Antibody for Western Blot
  • Host Rabbit
  • Target Species Human, Mouse, Rat
  • Conjugate Unconjugated
  • Catalog # PA527405
  • Size 100µL
Applications
Western Blot (1:500-1:3000)
Alternate names:
L-AIBAT; Gamma-amino-N-butyrate transaminase; GABA aminotransferase; (S)-3-amino-2-methylpropionate transaminase
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA527405 349.00
Check your price
  • ABAT Polyclonal Antibody for Western Blot
  • Host Rabbit
  • Target Species Bovine, Dog, Horse, Guinea pig, Goat, Human, Mouse, Rabbit, Rat
  • Conjugate Unconjugated
  • Catalog # PA543722
  • Size 100µl
Applications
Western Blot (0.2-1 µg/ml)
Alternate names:
L-AIBAT; Gamma-amino-N-butyrate transaminase; GABA aminotransferase; (S)-3-amino-2-methylpropionate transaminase
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA543722 309.00
Check your price
  • Phospho-STAT5 alpha (Ser726) Polyclonal Antibody for Western Blot, IHC and DB
  • Host Rabbit
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # PA512704
  • Size 400µL
Applications
Dot blot (1:500), Immunohistochemistry (1:50-1:100), Western Blot (1:1000)
Alternate names:
Signal transducer and activator of transcription 5A; STAT5A; STAT5;
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA512704 349.00
Check your price
  • TACR2 Polyclonal Antibody for IHC (P) and ELISA
  • Host Rabbit
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # PA533071
  • Size 50ug
Applications
ELISA (Assay-dependent), Immunohistochemistry (Paraffin) (40 µg/ml)
Alternate names:
Tachykinin receptor 2; Neurokinin A receptor; NK-2 receptor
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA533071 349.00
Check your price
  • TACR2 Polyclonal Antibody for IHC (P) and ELISA
  • Host Rabbit
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # PA533072
  • Size 50ug
Applications
ELISA (Assay-dependent), Immunohistochemistry (Paraffin) (20 µg/ml)
Alternate names:
Tachykinin receptor 2; Neurokinin A receptor; NK-2 receptor
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA533072 349.00
Check your price
  • TACR2 Polyclonal Antibody for IHC (P)
  • Host Rabbit
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # PA533968
  • Size 50ug
Applications
Immunohistochemistry (Paraffin) (15-30 µg/ml)
Alternate names:
Tachykinin receptor 2; Neurokinin A receptor; NK-2 receptor
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA533968 349.00
Check your price
  • TACR2 Polyclonal Antibody for IHC (P)
  • Host Rabbit
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # PA533969
  • Size 50ug
Applications
Immunohistochemistry (Paraffin) (5 µg/ml)
Alternate names:
Tachykinin receptor 2; Neurokinin A receptor; NK-2 receptor
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA533969 349.00
Check your price
  • TACR2 Polyclonal Antibody for IHC (P)
  • Host Rabbit
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # PA533967
  • Size 50ug
Applications
Immunohistochemistry (Paraffin) (5 µg/ml)
Alternate names:
Tachykinin receptor 2; Neurokinin A receptor; NK-2 receptor
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA533967 349.00
Check your price
  • Growth Hormone Polyclonal Antibody for IHC (P)
  • Host Rabbit
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # PA532448
  • Size 500µL
Applications
Immunohistochemistry (Paraffin) (1:200)
Alternate names:
Pituitary growth hormone; Growth hormone 1; Growth hormone
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA532448 349.00
Check your price
  • Growth Hormone Polyclonal Antibody for IHC (F) and IHC (P)
  • Host Rabbit
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # PA136094
  • Size 250µL
Applications
Immunohistochemistry (Paraffin, Frozen) (1:50)
Alternate names:
Pituitary growth hormone; Growth hormone 1; Growth hormone
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA136094 425.00
Check your price
  • Growth Hormone Polyclonal Antibody for Western Blot and IHC (P)
  • Host Rabbit
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # PA527360
  • Size 100µL
Applications
Immunohistochemistry (Paraffin) (1:100-1:1000), Western Blot (1:500-1:3000)
Alternate names:
Pituitary growth hormone; Growth hormone 1; Growth hormone
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA527360 349.00
Check your price
  • STAT5 beta Monoclonal Antibody for Western Blot
  • Host Mouse
  • Target Species Human, Mouse, Rat
  • Conjugate Unconjugated
  • Catalog # MA515665
  • Size 100µL
Applications
Western Blot (1/500-1/2000)
Alternate names:
STAT5; STAT5B
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA515665 349.00
Check your price
  • STAT5 alpha Polyclonal Antibody for Western Blot and IHC (P)
  • Host Rabbit
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # PA532602
  • Size 500µL
Applications
Immunohistochemistry (Paraffin) (1:200), Western Blot (1:25)
Alternate names:
MGF, STAT5
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA532602 275.00
Check your price