1 - 30 of 30 products displayed
  • Estrogen Receptor alpha Monoclonal Antibody for Western Blot, IF, ICC, IHC (F), IHC (P) and IP
  • Host Mouse
  • Target Species Human, Rat
  • Conjugate Unconjugated
  • Catalog # MA513304
  • Size 500 µL
Applications
Immunocytochemistry (1:50-1:200), Immunofluorescence (1:50-1:200), Immunohistochemistry (Paraffin, Frozen) (1:40-1:80), Immunoprecipitation (10 µl/mg lysate), Western Blot (1:100-1:1000)
Alternate names:
Nuclear receptor subfamily 3 group A member 1; Estradiol receptor; ER-alpha
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA513304 425.00
Check your price
  • Estrogen Receptor alpha Monoclonal Antibody for Western Blot, IHC (F), IHC (P) and IP
  • Host Mouse
  • Target Species Human, Rat
  • Conjugate Unconjugated
  • Catalog # MA127107
  • Size 250 µL
Applications
Immunohistochemistry (Frozen) (1:40-1:80), Immunohistochemistry (Paraffin) (1:40-1:80), Immunoprecipitation (10 µl/mg lysate), Western Blot (1:50-1:100)
Alternate names:
Nuclear receptor subfamily 3 group A member 1; Estradiol receptor; ER-alpha
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA127107 425.00
Check your price
  • Estrogen Receptor alpha Monoclonal Antibody for Western Blot, IHC (F), IHC (P) and IP
  • Host Mouse
  • Target Species Human, Rat
  • Conjugate Unconjugated
  • Catalog # MA180216
  • Size 100 µL
Applications
Immunohistochemistry (Frozen) (1:40-1:60), Immunohistochemistry (Paraffin) (1:40-1:80), Immunoprecipitation (10 µl/mg lysate), Western Blot (1:50-1:100)
Alternate names:
Nuclear receptor subfamily 3 group A member 1; Estradiol receptor; ER-alpha
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA180216 159.00
Check your price
  • Estrogen Receptor alpha Monoclonal Antibody, Biotin conjugate for IF, ICC, IP, GS and AP
  • Host Mouse
  • Target Species Bovine, Human, Mouse, Rabbit, Rat
  • Conjugate Biotin
  • Catalog # MA512325
  • Size 500 µL
Applications
Affinity Purification (Assay Dependent), Gel Shift (1mg/mL), Immunocytochemistry (1:500-1:2000), Immunofluorescence (1:500-1:2000), Immunoprecipitation (2 µg/ml)
Alternate names:
Nuclear receptor subfamily 3 group A member 1; Estradiol receptor; ER-alpha
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA512325 387.00
Check your price
  • Estrogen Receptor alpha Monoclonal Antibody for Western Blot, IF, ICC and IHC (P)
  • Host Mouse
  • Target Species Human, Rat
  • Conjugate Unconjugated
  • Catalog # MA514104
  • Size 500 µL
Applications
Immunocytochemistry (1:50-1:200), Immunofluorescence (1:50-1:200), Immunohistochemistry (Paraffin) (1:50-1:100), Western Blot (1:100-1:1000)
Alternate names:
Nuclear receptor subfamily 3 group A member 1; Estradiol receptor; ER-alpha
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA514104 550.00
Check your price
  • Estrogen Receptor alpha Monoclonal Antibody for Western Blot, IP and GS
  • Host Mouse
  • Target Species Bovine, Human, Mouse, Rabbit, Rat
  • Conjugate Unconjugated
  • Catalog # MA512328
  • Size 500 µL
Applications
Gel Shift (1mg/mL), Immunoprecipitation (2 µg/ml), Western Blot (1:100-1:400)
Alternate names:
Nuclear receptor subfamily 3 group A member 1; Estradiol receptor; ER-alpha
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA512328 309.00
Check your price
  • Estrogen Receptor alpha Monoclonal Antibody for Western Blot, IF, ICC and IHC (P)
  • Host Mouse
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # MA513191
  • Size 500 µL
Applications
Immunocytochemistry (1:500-1:1000), Immunofluorescence (1:500-1:1000), Immunohistochemistry (Paraffin) (0.4-0.8 µg/mL), Western Blot (0.5-1.0 µg/ml)
Alternate names:
Nuclear receptor subfamily 3 group A member 1; Estradiol receptor; ER-alpha
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA513191 550.00
Check your price
  • Estrogen Receptor alpha Monoclonal Antibody, Biotin conjugate for Western Blot, IP, ChIP and GS
  • Host Mouse
  • Target Species Bovine, Dog, Chicken, Hamster, Human, Mouse, Sheep, Pig, Rabbit, Rat
  • Conjugate Biotin
  • Catalog # MA513062
  • Size 500 µL
Applications
ChIP assay (Assay Dependent), Gel Shift (1mg/mL), Immunoprecipitation (2 µg/ml), Western Blot (1:1000-1:5000)
Alternate names:
Nuclear receptor subfamily 3 group A member 1; Estradiol receptor; ER-alpha
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA513062 399.00
Check your price
  • Estrogen Receptor alpha Polyclonal Antibody for Western Blot, IF, ICC and IHC (P)
  • Host Rabbit
  • Target Species Human, Pig, Rat
  • Conjugate Unconjugated
  • Catalog # PA1309
  • Size 50 µg
Applications
Immunocytochemistry (1:1000-1:4000), Immunofluorescence (1:1000-1:4000), Immunohistochemistry (Paraffin) (1-5 µg/ml), Western Blot (1 µg/ml)
Alternate names:
Nuclear receptor subfamily 3 group A member 1; Estradiol receptor; ER-alpha
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA1309 375.00
Check your price
  • Estrogen Receptor alpha Polyclonal Antibody for Western Blot, IF, ICC and IHC (F)
  • Host Rabbit
  • Target Species Human, Rat
  • Conjugate Unconjugated
  • Catalog # PA1308
  • Size 50 µg
Applications
Immunocytochemistry (1:1000-1:4000), Immunofluorescence (1:1000-1:4000), Immunohistochemistry (Frozen) (1-5 µg/ml), Western Blot (1 µg/ml)
Alternate names:
Nuclear receptor subfamily 3 group A member 1; Estradiol receptor; ER-alpha
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA1308 349.00
Check your price
  • Estrogen Receptor beta Monoclonal Antibody for Western Blot, IF, ICC, IHC (P), Flow and ChIP
  • Host Mouse
  • Target Species Human, Mouse
  • Conjugate Unconjugated
  • Catalog # MA123217
  • Size 100 µg
Applications
ChIP assay (Assay dependent), Flow Cytometry (Assay dependent), Immunocytochemistry (Assay dependent), Immunofluorescence (Assay dependent), Immunohistochemistry (Paraffin) (1-5 µg/ml), Western Blot (1-5 ug/ml)
Alternate names:
Nuclear receptor subfamily 3 group A member 2
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA123217 399.00
Check your price
  • Estrogen Receptor beta Polyclonal Antibody for Western Blot, IHC (F) and IHC (P)
  • Host Rabbit
  • Target Species Human, Rat
  • Conjugate Unconjugated
  • Catalog # PA1313
  • Size 50 µg
Applications
Immunohistochemistry (Paraffin, Frozen) (2-5 µg/ml), Western Blot (1 µg/ml)
Alternate names:
Nuclear receptor subfamily 3 group A member 2
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA1313 349.00
Check your price
  • Estrogen Receptor alpha Monoclonal Antibody for Western Blot
  • Host Mouse
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # MA515402
  • Size 100 µL
Applications
Western Blot (1/500-1/2000)
Alternate names:
Nuclear receptor subfamily 3 group A member 1; Estradiol receptor; ER-alpha
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA515402 349.00
Check your price
  • Estrogen Receptor alpha Monoclonal Antibody for Western Blot and IHC
  • Host Mouse
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # MA515268
  • Size 100 ug
Applications
Immunohistochemistry (1/200-1/1000), Western Blot (1/500-1/2000)
Alternate names:
Nuclear receptor subfamily 3 group A member 1; Estradiol receptor; ER-alpha
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA515268 349.00
Check your price
  • Estrogen Receptor alpha Monoclonal Antibody for Western Blot
  • Host Mouse
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # MA515454
  • Size 100 µL
Applications
Western Blot (1/500-1/2000)
Alternate names:
Nuclear receptor subfamily 3 group A member 1; Estradiol receptor; ER-alpha
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA515454 349.00
Check your price
  • Estrogen Receptor alpha Monoclonal Antibody for Western Blot
  • Host Mouse
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # MA515344
  • Size 100 µL
Applications
Western Blot (1/500-1/2000)
Alternate names:
Nuclear receptor subfamily 3 group A member 1; Estradiol receptor; ER-alpha
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA515344 349.00
Check your price
  • Estrogen Receptor alpha Monoclonal Antibody for Western Blot, IF, ICC and IHC
  • Host Mouse
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # MA515689
  • Size 100 µL
Applications
Immunocytochemistry (1:200-1:1000), Immunofluorescence (1/200-1/1000), Immunohistochemistry (1/200-1/1000), Western Blot (1/500-1/2000)
Alternate names:
Nuclear receptor subfamily 3 group A member 1; Estradiol receptor; ER-alpha
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA515689 349.00
Check your price
  • Estrogen Receptor alpha Monoclonal Antibody for Western Blot, IHC, IP and ELISA
  • Host Mouse
  • Target Species Human, Rat
  • Conjugate Unconjugated
  • Catalog # 417000
  • Size 100 µg
Applications
ELISA (Assay Dependent), Immunohistochemistry (Assay Dependent), Immunoprecipitation (Assay Dependent), Western Blot (Assay Dependent)
Alternate names:
Nuclear receptor subfamily 3 group A member 1; Estradiol receptor; ER-alpha
Catalog # Size Price (AUD) Your Price (AUD) Qty
417000 433.00
Check your price
  • Estrogen Receptor beta Polyclonal Antibody for Western Blot, IF, ICC, IHC (F), IHC (P) and GS
  • Host Rabbit
  • Target Species Human, Mouse, Non-human primate, Sheep, Rat
  • Conjugate Unconjugated
  • Catalog # PA1311
  • Size 50 µg
Applications
Gel Shift (1-5 µl), Immunocytochemistry (2-4 µg/ml), Immunofluorescence (0.5 µg/ml), Immunohistochemistry (Paraffin, Frozen) (1-2 µg/ml), Western Blot (1-2 µg/ml)
Alternate names:
Nuclear receptor subfamily 3 group A member 2
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA1311 375.00
Check your price
  • Estrogen Receptor beta Polyclonal Antibody for Western Blot, IF, ICC and IHC
  • Host Rabbit
  • Target Species Human, Mouse, Rat
  • Conjugate Unconjugated
  • Catalog # PA1312
  • Size 50 µg
Applications
Immunocytochemistry (1:50-1:200), Immunofluorescence (1:50-1:200), Immunohistochemistry (1-2 µg/ml), Western Blot (2 µg/ml)
Alternate names:
Nuclear receptor subfamily 3 group A member 2
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA1312 349.00
Check your price
  • Estrogen Receptor beta Polyclonal Antibody for Western Blot, IF, ICC, IHC (F) and IHC (P)
  • Host Rabbit
  • Target Species Human, Mouse, Rat
  • Conjugate Unconjugated
  • Catalog # PA1310B
  • Size 100 µg
Applications
Immunocytochemistry (1:100-1:1000), Immunofluorescence (1:100-1:1000), Immunohistochemistry (Paraffin, Frozen) (1:50), Western Blot (1 µg/ml)
Alternate names:
Nuclear receptor subfamily 3 group A member 2
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA1310B 375.00
Check your price
  • TGF beta Receptor I Polyclonal Antibody for Western Blot, IF, ICC, IHC (P) and Flow
  • Host Rabbit
  • Target Species Human, Rat
  • Conjugate
  • Catalog # PA527785
  • Size 100µL
Applications
Immunocytochemistry (1:100-1:1000), Immunofluorescence (1:100-1:1000), Immunohistochemistry (Paraffin) (1:100-1:1000), Western Blot (1:1000-1:10,000)
Alternate names:
Transforming growth factor-beta receptor type I; TGF-beta type I receptor; Serine/threonine-protein kinase receptor R4; Activin receptor-like kinase 5; Activin A receptor type II-like protein kinase of 53kD
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA527785 349.00
Check your price
  • Progesterone Receptor Monoclonal Antibody for Western Blot, IF and IHC (P)
  • Host Rabbit
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # MA514505
  • Size 500 µL
Applications
Immunofluorescence (1-2 µg/ml), Immunohistochemistry (Paraffin) (1:100-1:400), Western Blot (1:1000-1:2000)
Alternate names:
Nuclear receptor subfamily 3 group C member 3
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA514505 700.00
Check your price
  • Progesterone Receptor Monoclonal Antibody for IHC (P)
  • Host Rabbit
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # MA516393
  • Size 500 µL
Applications
Immunohistochemistry (Paraffin) (1:400)
Alternate names:
Nuclear receptor subfamily 3 group C member 3
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA516393 700.00
Check your price
  • Prohibitin Monoclonal Antibody for Western Blot, IF and IHC (P)
  • Host Mouse
  • Target Species Chicken, Human, Mouse, Pig, Rat
  • Conjugate Unconjugated
  • Catalog # MA512858
  • Size 500 µL
Applications
Immunofluorescence (Assay Dependent), Immunohistochemistry (Paraffin) (2-4 µg/ml), Western Blot (0.5-1.0 µg/ml)
Alternate names:
Repressor of estrogen receptor activity; D-prohibitin; B-cell receptor-associated protein BAP37; B-cell receptor-associated protein 32
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA512858 349.00
Check your price
  • Prohibitin Monoclonal Antibody, Biotin conjugate for Western Blot, IF and IHC (P)
  • Host Mouse
  • Target Species Chicken, Human, Mouse, Pig, Rat
  • Conjugate Biotin
  • Catalog # MA512855
  • Size 500 µL
Applications
Immunofluorescence (Assay Dependent), Immunohistochemistry (Paraffin) (2-4 µg/ml), Western Blot (0.5-1.0 µg/ml)
Alternate names:
Repressor of estrogen receptor activity; D-prohibitin; B-cell receptor-associated protein BAP37; B-cell receptor-associated protein 32
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA512855 399.00
Check your price
  • HCC1 Polyclonal Antibody for Western Blot
  • Host Rabbit
  • Target Species Human, Mouse
  • Conjugate Unconjugated
  • Catalog # PA531103
  • Size 100 µL
Applications
Western Blot (1:500-1:3000)
Alternate names:
Splicing factor HCC1; RNA-binding region-containing protein 2; RNA-binding motif protein 39; Hepatocellular carcinoma protein 1; CAPER alpha; Transcription coactivator CAPER; Coactivator of activating protein 1 and estrogen receptors
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA531103 349.00
Check your price
  • Progesterone Receptor Monoclonal Antibody for IHC (P)
  • Host Rabbit
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # MA516394
  • Size 500 µL
Applications
Immunohistochemistry (Paraffin) (1:400)
Alternate names:
Nuclear receptor subfamily 3 group C member 3
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA516394 700.00
Check your price
  • ESRRA Polyclonal Antibody for Western Blot, IP and ChIP
  • Host Rabbit
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # PA528390
  • Size 100 µL
Applications
ChIP assay (Assay-dependent), Immunoprecipitation (1:100-1:500), Western Blot (1:500-1:3000)
Alternate names:
Nuclear receptor subfamily 3 group B member 1; Estrogen-related receptor alpha; Estrogen receptor-like 1
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA528390 349.00
Check your price
  • ESRRG Polyclonal Antibody for Western Blot
  • Host Rabbit
  • Target Species Human, Mouse, Rat
  • Conjugate Unconjugated
  • Catalog # PA527755
  • Size 100 µL
Applications
Western Blot (1:500-1:3000)
Alternate names:
Nuclear receptor subfamily 3 group B member 3; Estrogen receptor-related protein 3; ERR gamma-2
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA527755 349.00
Check your price