1 - 7 of 7 products displayed
  • MMP9 (human) for Western Blot and Ctrl
  • Host E. Coli
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # RP75654
  • Size 100µg
Applications
Control (Assay Dependent), Western Blot (Assay Dependent)
Alternate names:
Gelatinase B; 92 kDa type IV collagenase; 92 kDa gelatinase
Catalog # Size Price (AUD) Your Price (AUD) Qty
RP75654 598.00
Check your price
  • MMP9 Monoclonal Antibody for Western Blot and IP
  • Host Mouse
  • Target Species Human, Rat
  • Conjugate Unconjugated
  • Catalog # MS817PABX
  • Size 200ug
Applications
Immunoprecipitation (Assay Dependent), Western Blot (Assay Dependent)
Alternate names:
Gelatinase B; 92 kDa type IV collagenase; 92 kDa gelatinase
Catalog # Size Price (AUD) Your Price (AUD) Qty
MS817PABX 499.00
Check your price
  • MMP9 Monoclonal Antibody for Western Blot, IF, ICC, IHC and Flow
  • Host Mouse
  • Target Species Human, Mouse
  • Conjugate Unconjugated
  • Catalog # MA515886
  • Size 100µL
Applications
Flow Cytometry (1/200-1/400), Immunocytochemistry (1:200-1:1000), Immunofluorescence (1/200-1/1000), Immunohistochemistry (1/200-1/1000), Western Blot (1/500-1/2000)
Alternate names:
Gelatinase B; 92 kDa type IV collagenase; 92 kDa gelatinase
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA515886 349.00
Check your price
  • MMP9 Monoclonal Antibody for Western Blot and Flow
  • Host Mouse
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # MA515888
  • Size 100µL
Applications
Flow Cytometry (1/200-1/400), Western Blot (1/500-1/2000)
Alternate names:
Gelatinase B; 92 kDa type IV collagenase; 92 kDa gelatinase
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA515888 349.00
Check your price
  • MMP9 Polyclonal Antibody for Western Blot, IF, ICC and IHC (P)
  • Host Rabbit
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # PA527191
  • Size 100µL
Applications
Immunocytochemistry (1:100-1:1000), Immunofluorescence (1:100-1:1000), Immunohistochemistry (Paraffin) (1:100-1:1000), Western Blot (1:500-1:3000)
Alternate names:
Gelatinase B; 92 kDa type IV collagenase; 92 kDa gelatinase
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA527191 349.00
Check your price
  • MMP9 (human) for Western Blot and Ctrl
  • Host E. Coli
  • Target Species
  • Conjugate Unconjugated
  • Catalog # RP75653
  • Size 10µg
Applications
Control (Assay Dependent), Western Blot (Assay Dependent)
Alternate names:
Catalog # Size Price (AUD) Your Price (AUD) Qty
RP75653 209.00
Check your price
  • MMP9 (human) for Western Blot and Ctrl
  • Host E. Coli
  • Target Species Not Applicable
  • Conjugate Unconjugated
  • Catalog # RP75655
  • Size 1mg
Applications
Control (Assay Dependent), Western Blot (Assay Dependent)
Alternate names:
Catalog # Size Price (AUD) Your Price (AUD) Qty
RP75655 2,900.00
Check your price