1 - 10 of 10 products displayed
  • TNFRSF14 Monoclonal Antibody for IF and Flow
  • Host Mouse
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # MA513986
  • Size 500 µL
Applications
Flow Cytometry (Assay Dependent), Immunofluorescence (Assay Dependent), Inhibition Assays (Assay Dependent)
Alternate names:
Tumor necrosis factor receptor-like 2; Herpes virus entry mediator A
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA513986 425.00
Check your price
  • TNFRSF14 Monoclonal Antibody for IF, ICC, Flow and IP
  • Host Mouse
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # MA125958
  • Size 250 µL
Applications
Flow Cytometry (Assay Dependent), Immunocytochemistry (Assay-dependent), Immunofluorescence (Assay Dependent), Immunoprecipitation (2 µg/mg of protein lysate)
Alternate names:
Tumor necrosis factor receptor-like 2; Herpes virus entry mediator A
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA125958 425.00
Check your price
  • TNFRSF14 Polyclonal Antibody for Western Blot, IF, ICC and IHC
  • Host Rabbit
  • Target Species Human, Mouse
  • Conjugate Unconjugated
  • Catalog # PA520237
  • Size 100 ug
Applications
Immunocytochemistry (10 ug/ml), Immunofluorescence (10 ug/ml), Immunohistochemistry (1 µg/ml), Western Blot (1-2 ug/ml)
Alternate names:
Tumor necrosis factor receptor-like 2; Herpes virus entry mediator A
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA520237 349.00
Check your price
  • TNFRSF14 Polyclonal Antibody for Western Blot, IF and ICC
  • Host Rabbit
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # PA520236
  • Size 100 ug
Applications
Immunocytochemistry (10 µg/ml), Immunofluorescence (20 ug/ml), Western Blot (2 µg/ml)
Alternate names:
Tumor necrosis factor receptor-like 2; Herpes virus entry mediator A
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA520236 349.00
Check your price
  • TNFRSF14 Polyclonal Antibody for Western Blot, IHC and Flow
  • Host Rabbit
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # PA526103
  • Size 400 µL
Applications
Flow Cytometry (1:10-1:50), Immunohistochemistry (1:50-1:100), Western Blot (1:1000)
Alternate names:
Tumor necrosis factor receptor-like 2; Herpes virus entry mediator A
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA526103 349.00
Check your price
  • TNFRSF14 Polyclonal Antibody for Western Blot
  • Host Goat
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # PA518451
  • Size 100 ug
Applications
Western Blot (0.03-0.1 ug/ml)
Alternate names:
Tumor necrosis factor receptor-like 2; Herpes virus entry mediator A
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA518451 349.00
Check your price
  • TNFRSF14 Polyclonal Antibody for Western Blot
  • Host Rabbit
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # PA529780
  • Size 100 µL
Applications
Western Blot (1:500-1:3000)
Alternate names:
Tumor necrosis factor receptor-like 2; Herpes virus entry mediator A
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA529780 349.00
Check your price
  • TNFRSF14 Monoclonal Antibody for IF, ICC, Flow and IP
  • Host Rat
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # MA125844
  • Size 50 µg
Applications
Flow Cytometry (1 µg/ml), Immunocytochemistry (1:1000), Immunofluorescence (1:1000), Immunoprecipitation (1:200)
Alternate names:
Tumor necrosis factor receptor-like 2; Herpes virus entry mediator A
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA125844 425.00
Check your price
  • LIGHT Monoclonal Antibody for Western Blot
  • Host Rat
  • Target Species Mouse
  • Conjugate Unconjugated
  • Catalog # MA523961
  • Size 100 µg
Applications
Western Blot (1 µg/ml)
Alternate names:
TNFSF14, tumor necrosis factor (ligand) superfamily, member 14, CD258, HVEML, LIGHT, LTg, TR2, delta transmembrane LIGHT, herpesvirus entry mediator A, ligand for herpesvirus entry mediator, tumor necrosis factor ligand superfamily, member 14, tumor necrosis factor receptor-like 2, tumor necrosis factor superfamily member LIGHT
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA523961 275.00
Check your price
  • LIGHT Polyclonal Antibody for Western Blot
  • Host Goat
  • Target Species Mouse
  • Conjugate Unconjugated
  • Catalog # PA547203
  • Size 100 µg
Applications
Western Blot (0.1 µg/ml)
Alternate names:
TNFSF14, tumor necrosis factor (ligand) superfamily, member 14, CD258, HVEML, LIGHT, LTg, TR2, delta transmembrane LIGHT, herpesvirus entry mediator A, ligand for herpesvirus entry mediator, tumor necrosis factor ligand superfamily, member 14, tumor necrosis factor receptor-like 2, tumor necrosis factor superfamily member LIGHT
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA547203 349.00
Check your price