1 - 27 of 27 products displayed
  • IA2 Monoclonal Antibody for Western Blot and ELISA
  • Host Mouse
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # MA517229
  • Size 100 µL
Applications
ELISA (Assay Dependent), Western Blot (2 µg/ml)
Alternate names:
PTP IA-2; Islet cell autoantigen 3; Islet cell antigen 512
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA517229 349.00
Check your price
  • IA2 Polyclonal Antibody for Western Blot
  • Host Chicken
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # PA127627
  • Size 50 µg
Applications
Western Blot (1:2000)
Alternate names:
PTP IA-2; Islet cell autoantigen 3; Islet cell antigen 512
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA127627 425.00
Check your price
  • IA2 Polyclonal Antibody for Western Blot
  • Host Goat
  • Target Species Mouse
  • Conjugate Unconjugated
  • Catalog # PA518906
  • Size 100 ug
Applications
Western Blot (0.5-1.5 ug/ml)
Alternate names:
IA-2 ; protein tyrosine phosphatase, receptor type, N ; FLJ16131 ; IA-2/PTP; IA2 ; ICA512 ; R-PTP-N; islet antigen 2 ; islet cell antigen 512 ; islet cell autoantigen 3 ; protein tyrosine phosphatase-like N
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA518906 349.00
Check your price
  • I-Ab/I-Ad Monoclonal Antibody, Biotin conjugate for Flow
  • Host Mouse
  • Target Species Mouse
  • Conjugate Biotin
  • Catalog # MA517775
  • Size 100 ug
Applications
Flow Cytometry (0.1 ug/10^6 cells)
Alternate names:
H-2 class II histocompatibility antigen A beta chain, H2-AB1, H2-iabeta
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA517775 159.00
Check your price
  • I-Ab/I-Ad Monoclonal Antibody, FITC conjugate for Flow
  • Host Mouse
  • Target Species Mouse
  • Conjugate FITC
  • Catalog # MA517776
  • Size 100 ug
Applications
Flow Cytometry (0.5 ug/10^6 cells)
Alternate names:
H-2 class II histocompatibility antigen A beta chain, H2-AB1, H2-iabeta
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA517776 159.00
Check your price
  • I-Ab/I-Ad Monoclonal Antibody, PE conjugate for Flow
  • Host Mouse
  • Target Species Mouse
  • Conjugate PE
  • Catalog # MA517777
  • Size 50 ug
Applications
Flow Cytometry (0.5 ug/10^6 cells)
Alternate names:
H-2 class II histocompatibility antigen A beta chain, H2-AB1, H2-iabeta
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA517777 139.00
Check your price
  • I-Ab Monoclonal Antibody, Biotin conjugate for Flow
  • Host Mouse
  • Target Species Mouse
  • Conjugate Biotin
  • Catalog # MA517759
  • Size 100 ug
Applications
Flow Cytometry (2.0 ug/10^6 cells)
Alternate names:
H-2 class II histocompatibility antigen A beta chain, H2-AB1, H2-iabeta
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA517759 159.00
Check your price
  • I-Ad Monoclonal Antibody, Biotin conjugate for Flow
  • Host Mouse
  • Target Species Mouse
  • Conjugate Biotin
  • Catalog # MA517778
  • Size 100 ug
Applications
Flow Cytometry (0.1 ug/10^6 cells)
Alternate names:
H-2 class II histocompatibility antigen A beta chain, H2-AB1, H2-iabeta
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA517778 159.00
Check your price
  • I-Ab Monoclonal Antibody, FITC conjugate for Flow
  • Host Mouse
  • Target Species Mouse
  • Conjugate FITC
  • Catalog # MA517760
  • Size 100 ug
Applications
Flow Cytometry (2.0 ug/10^6 cells)
Alternate names:
H-2 class II histocompatibility antigen A beta chain, H2-AB1, H2-iabeta
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA517760 159.00
Check your price
  • I-Ad Monoclonal Antibody, FITC conjugate for Flow
  • Host Mouse
  • Target Species Mouse
  • Conjugate FITC
  • Catalog # MA517779
  • Size 100 ug
Applications
Flow Cytometry (0.1 ug/10^6 cells)
Alternate names:
H-2 class II histocompatibility antigen A beta chain, H2-AB1, H2-iabeta
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA517779 159.00
Check your price
  • I-Ab Monoclonal Antibody, PE conjugate for Flow
  • Host Mouse
  • Target Species Mouse
  • Conjugate PE
  • Catalog # MA517758
  • Size 50 ug
Applications
Flow Cytometry (0.2 ug/10^6 cells)
Alternate names:
H-2 class II histocompatibility antigen A beta chain, H2-AB1, H2-iabeta
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA517758 139.00
Check your price
  • I-Ad Monoclonal Antibody, PE conjugate for Flow
  • Host Mouse
  • Target Species Mouse
  • Conjugate PE
  • Catalog # MA517780
  • Size 50 ug
Applications
Flow Cytometry (0.1 ug/10^6 cells)
Alternate names:
H-2 class II histocompatibility antigen A beta chain, H2-AB1, H2-iabeta
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA517780 139.00
Check your price
  • I-Ab Monoclonal Antibody for Flow
  • Host Mouse
  • Target Species Mouse
  • Conjugate Unconjugated
  • Catalog # MA170065
  • Size 250 ug
Applications
Flow Cytometry (1 ug/1 million cells)
Alternate names:
H-2Eb, H2Eb, Ia-4, Ia4, MGC163794, MGC163796
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA170065 275.00
Check your price
  • Blood Group H Type 2 Monoclonal Antibody for ICC, IHC (F), IHC (P) and ELISA
  • Host Mouse
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # MA135386
  • Size 100 µg
Applications
ELISA (Assay Dependent), Immunocytochemistry (Assay Dependent), Immunohistochemistry (Paraffin, Frozen) (Assay Dependent)
Alternate names:
AI845868, Abeta, H-2Ab, H2-Ab, I-A, I-Abeta, IAb, Ia-2, Ia2, Rmcs1
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA135386 349.00
Check your price
  • MHC Class II (I-A / I-E) Monoclonal Antibody for Flow
  • Host Rat
  • Target Species Mouse
  • Conjugate
  • Catalog # A14901
  • Size 25µg
Applications
Flow Cytometry (Assay Dependent)
Alternate names:
H-2 class II histocompatibility antigen A beta chain, H2-AB1, H2-iabeta
Catalog # Size Price (AUD) Your Price (AUD) Qty
A14901 103.00
Check your price
  • I-Ab Monoclonal Antibody for Flow
  • Host Mouse
  • Target Species Mouse
  • Conjugate
  • Catalog # A14909
  • Size 25µg
Applications
Flow Cytometry (Assay Dependent)
Alternate names:
H-2 class II histocompatibility antigen A beta chain, H2-AB1, H2-iabeta
Catalog # Size Price (AUD) Your Price (AUD) Qty
A14909 130.00
Check your price
  • I-Ab Monoclonal Antibody for Flow
  • Host Mouse
  • Target Species Mouse
  • Conjugate
  • Catalog # A14908
  • Size 25µg
Applications
Flow Cytometry (Assay Dependent)
Alternate names:
H-2 class II histocompatibility antigen A beta chain, H2-AB1, H2-iabeta
Catalog # Size Price (AUD) Your Price (AUD) Qty
A14908 104.00
Check your price
  • I-Ab Monoclonal Antibody, Biotin conjugate for Flow
  • Host Mouse
  • Target Species Mouse
  • Conjugate Biotin
  • Catalog # MA517756
  • Size 100 ug
Applications
Flow Cytometry (Assay Dependent)
Alternate names:
H-2 class II histocompatibility antigen A beta chain, H2-AB1, H2-iabeta
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA517756 159.00
Check your price
  • I-Ab Monoclonal Antibody, FITC conjugate for Flow
  • Host Mouse
  • Target Species Mouse
  • Conjugate FITC
  • Catalog # MA517757
  • Size 100 ug
Applications
Flow Cytometry (Assay Dependent)
Alternate names:
H-2 class II histocompatibility antigen A beta chain, H2-AB1, H2-iabeta
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA517757 159.00
Check your price
  • I-A/I-E Monoclonal Antibody, FITC conjugate for Flow
  • Host Rat
  • Target Species Mouse
  • Conjugate FITC
  • Catalog # MA110403
  • Size 100 ug
Applications
Flow Cytometry (4 ug/ml)
Alternate names:
I-Ab, I-Ad, I-Aq, I-Ed, I-Ek, MHC class 2, MHC class II glygoproteins
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA110403 225.00
Check your price
  • MHC Class II (I-A / I-E) Monoclonal Antibody for Flow
  • Host Rat
  • Target Species Mouse
  • Conjugate
  • Catalog # A14902
  • Size 25µg
Applications
Flow Cytometry (Assay Dependent)
Alternate names:
H-2 class II histocompatibility antigen A beta chain, H2-AB1, H2-iabeta
Catalog # Size Price (AUD) Your Price (AUD) Qty
A14902 125.00
Check your price
  • I-Ab Monoclonal Antibody for IHC (F) and Flow
  • Host Rat
  • Target Species Mouse
  • Conjugate Unconjugated
  • Catalog # MA181017
  • Size 2 mL
Applications
Flow Cytometry (1:5), Immunohistochemistry (Frozen) (Neat)
Alternate names:
DADB-249P12.2, CELIAC1, HLA-DQB, IDDM1
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA181017 249.00
Check your price
  • I-Ab Monoclonal Antibody for Flow
  • Host Mouse
  • Target Species Mouse
  • Conjugate
  • Catalog # A14910
  • Size 25µg
Applications
Flow Cytometry (Assay Dependent)
Alternate names:
H-2 class II histocompatibility antigen A beta chain, H2-AB1, H2-iabeta
Catalog # Size Price (AUD) Your Price (AUD) Qty
A14910 113.00
Check your price
  • I-Ab Monoclonal Antibody for Flow
  • Host Mouse
  • Target Species Mouse
  • Conjugate
  • Catalog # A14907
  • Size 25µg
Applications
Flow Cytometry (Assay Dependent)
Alternate names:
H-2 class II histocompatibility antigen A beta chain, H2-AB1, H2-iabeta
Catalog # Size Price (AUD) Your Price (AUD) Qty
A14907 144.00
Check your price
  • I-Ab Monoclonal Antibody, PE conjugate for Flow
  • Host Mouse
  • Target Species Mouse
  • Conjugate PE
  • Catalog # A14747
  • Size 25 µg
Applications
Flow Cytometry (Assay Dependent)
Alternate names:
H-2 class II histocompatibility antigen A beta chain, H2-AB1, H2-iabeta
Catalog # Size Price (AUD) Your Price (AUD) Qty
A14747 103.00
Check your price
  • I-Ab Monoclonal Antibody for Flow
  • Host Mouse
  • Target Species Mouse
  • Conjugate Unconjugated
  • Catalog # MA170064
  • Size 250 ug
Applications
Flow Cytometry (1 ug/1 million cells)
Alternate names:
H-2Eb, H2Eb, Ia-4, Ia4, MGC163794, MGC163796
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA170064 275.00
Check your price
  • I-Ab Monoclonal Antibody for IHC (F) and Flow
  • Host Rat
  • Target Species Mouse, Rat
  • Conjugate Unconjugated
  • Catalog # MA516913
  • Size 5 mL
Applications
Flow Cytometry (Neat-1:4), Immunohistochemistry (Frozen) (Assay Dependent)
Alternate names:
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA516913 159.00
Check your price