1 - 7 of 7 products displayed
  • IDH1 Polyclonal Antibody for Western Blot, IF, ICC, IHC (P) and Flow
  • Host Rabbit
  • Target Species Human, Mouse
  • Conjugate Unconjugated
  • Catalog # PA514358
  • Size 400 µL
Applications
Flow Cytometry (1:10-1:50), Immunocytochemistry (1:10-1:50), Immunofluorescence (1:10-1:50), Immunohistochemistry (1:50-1:100), Western Blot (1:1000)
Alternate names:
Oxalosuccinate decarboxylase; NADP(+)-specific ICDH; IDP; Cytosolic NADP-isocitrate dehydrogenase
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA514358 349.00
Check your price
  • IDH1 Polyclonal Antibody for Western Blot, IF, ICC, IHC and Flow
  • Host Rabbit
  • Target Species Human, Mouse
  • Conjugate Unconjugated
  • Catalog # PA514357
  • Size 400 µL
Applications
Flow Cytometry (1:10-1:50), Immunocytochemistry (1:10-1:50), Immunofluorescence (1:10-1:50), Immunohistochemistry (1:50-1:100), Western Blot (1:1000)
Alternate names:
Oxalosuccinate decarboxylase; NADP(+)-specific ICDH; IDP; Cytosolic NADP-isocitrate dehydrogenase
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA514357 349.00
Check your price
  • IDH1 Polyclonal Antibody for Western Blot
  • Host Rabbit
  • Target Species Human, Mouse, Rat
  • Conjugate Unconjugated
  • Catalog # PA520976
  • Size 100 ug
Applications
Western Blot (1-2 ug/ml)
Alternate names:
Oxalosuccinate decarboxylase; NADP(+)-specific ICDH; IDP; Cytosolic NADP-isocitrate dehydrogenase
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA520976 349.00
Check your price
  • IDH1 Polyclonal Antibody for Western Blot, IHC (P) and IP
  • Host Rabbit
  • Target Species Human, Mouse
  • Conjugate Unconjugated
  • Catalog # PA528206
  • Size 100 µL
Applications
Immunohistochemistry (Paraffin) (1:100-1:1000), Immunoprecipitation (1:100-1:1000), Western Blot (1:500-1:3000)
Alternate names:
Oxalosuccinate decarboxylase; NADP(+)-specific ICDH; IDP; Cytosolic NADP-isocitrate dehydrogenase
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA528206 349.00
Check your price
  • IDH1 Polyclonal Antibody for Western Blot
  • Host Goat
  • Target Species Human, Mouse, Rat
  • Conjugate Unconjugated
  • Catalog # PA519414
  • Size 100 ug
Applications
Western Blot (0.1-0.3 ug/ml)
Alternate names:
Oxalosuccinate decarboxylase; NADP(+)-specific ICDH; IDP; Cytosolic NADP-isocitrate dehydrogenase
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA519414 349.00
Check your price
  • IDH1 Polyclonal Antibody for Western Blot
  • Host Goat
  • Target Species Human, Mouse
  • Conjugate Unconjugated
  • Catalog # PA519413
  • Size 100 ug
Applications
Western Blot (0.3-1 ug/ml)
Alternate names:
Oxalosuccinate decarboxylase; NADP(+)-specific ICDH; IDP; Cytosolic NADP-isocitrate dehydrogenase
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA519413 349.00
Check your price
  • IDH2 Polyclonal Antibody for Western Blot, IF, ICC and IHC
  • Host Rabbit
  • Target Species Human, Mouse, Rat
  • Conjugate Unconjugated
  • Catalog # PA520977
  • Size 100 ug
Applications
Immunocytochemistry (20 ug/ml), Immunofluorescence (20 ug/ml), Immunohistochemistry (5 ug/ml), Western Blot (1-2 ug/ml)
Alternate names:
Oxalosuccinate decarboxylase; NADP(+)-specific ICDH; IDP; ICD-M
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA520977 349.00
Check your price