1 - 3 of 3 products displayed
  • IDUA Polyclonal Antibody for Western Blot, IHC and Flow
  • Host Rabbit
  • Target Species Human
  • Conjugate
  • Catalog # PA525262
  • Size 400µL
Applications
Flow Cytometry (1:10-1:50), Immunohistochemistry (1:50-1:100), Western Blot (1:1000)
Alternate names:
IDA, MPS1, alpha-L-iduronidase
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA525262 Not Orderable Online
Check your price
  • IDUA Polyclonal Antibody for Western Blot, IF, ICC and IP
  • Host Sheep
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # PA547583
  • Size 100µg
Applications
Immunocytochemistry (5-15 µg/ml), Immunofluorescence (5-15 µg/ml), Immunoprecipitation (25 µg/ml), Western Blot (0.1 µg/ml)
Alternate names:
IDA, MPS1, alpha-L-iduronidase
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA547583 349.00
Check your price
  • IDUA Polyclonal Antibody for Western Blot
  • Host Rabbit
  • Target Species Bovine, Dog, Horse, Guinea pig, Human, Mouse, Rat
  • Conjugate Unconjugated
  • Catalog # PA545483
  • Size 100µl
Applications
Western Blot (1 µg/ml )
Alternate names:
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA545483 309.00
Check your price