1 - 4 of 4 products displayed
  • Goat anti-Pig IgA Secondary Antibody, HRP conjugate for ELISA
  • Host Goat
  • Target Species Pig
  • Conjugate HRP
  • Catalog # PA184625
  • Size 1mg
Applications
ELISA (1/10,000-1/100,000)
Alternate names:
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA184625 423.00
Check your price
  • Pig IgA (Secretory chain) Monoclonal Secondary Antibody for IHC (F), ELISA
  • Host Mouse
  • Target Species Pig
  • Conjugate
  • Catalog # MA180544
  • Size 2mL
Applications
ELISA (Assay Dependent), Immunohistochemistry (Frozen) (Assay Dependent)
Alternate names:
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA180544 Not Orderable Online
Check your price
  • Goat anti-Pig IgA Secondary Antibody, FITC conjugate for IHC (F) and Flow
  • Host Goat
  • Target Species Pig
  • Conjugate FITC
  • Catalog # PA184624
  • Size 1mg
Applications
Flow Cytometry (Assay Dependent), Immunohistochemistry (Frozen) (1:50-1:500)
Alternate names:
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA184624 364.00
Check your price
  • TGF beta-1 Monoclonal Antibody for Western Blot, IHC (P), Flow, ELISA and Neu
  • Host Mouse
  • Target Species Mouse
  • Conjugate Unconjugated
  • Catalog # MA523702
  • Size 500µg
Applications
ELISA (0.1-0.4 µg/ml (Detection), 2-8 µg/ml (Capture)), Flow Cytometry (2.5 µg/10^6 cells), Immunohistochemistry (Paraffin) (Assay-Dependent), Neutralization (0.3-1.0 µg/ml), Western Blot (1 µg/ml)
Alternate names:
TGFB, TGFb-1, DPD1, Transforming Growth Factor, Transforming Growth Factor Beta-1, TGF-beta-1, CED
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA523702 375.00
Check your price