1 - 30 of 30 products displayed
  • IGF1 Monoclonal Antibody for Western Blot and ELISA
  • Host Mouse
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # MA1088
  • Size 200ug
Applications
ELISA (1ug/ml), Western Blot (1:500-1:2000)
Alternate names:
Somatomedin-C; Mechano growth factor
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA1088 309.00
Check your price
  • IGF1 Monoclonal Antibody for Western Blot, ELISA and Neu
  • Host Hamster
  • Target Species Mouse
  • Conjugate Unconjugated
  • Catalog # MA523772
  • Size 500µg
Applications
ELISA (0.1-0.4 µg/ml (Detection), 2-8 µg/ml (Capture)), Neutralization (0.3-1.5 µg/ml), Western Blot (1 µg/ml)
Alternate names:
Somatomedin
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA523772 349.00
Check your price
  • IGF1 Monoclonal Antibody for IHC (F), IHC (P), ELISA and RIA
  • Host Mouse
  • Target Species Human, Mouse, Rat
  • Conjugate Unconjugated
  • Catalog # MA518035
  • Size 100µL
Applications
ELISA (Assay-dependent), Immunohistochemistry (Frozen) (Assay-dependent), Immunohistochemistry (Paraffin) (1 ug/ml), Radioimmune Assays (Assay-dependent)
Alternate names:
Somatomedin; Somatomedin-C; Mechano growth factor
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA518035 425.00
Check your price
  • IGF1 Monoclonal Antibody for Western Blot
  • Host Mouse
  • Target Species Human, Mouse, Rat
  • Conjugate Unconjugated
  • Catalog # MA512247
  • Size 500µL
Applications
Western Blot (1-2 µg/ml)
Alternate names:
Somatomedin; Somatomedin-C; Mechano growth factor
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA512247 349.00
Check your price
  • IGF1 Monoclonal Antibody for Western Blot
  • Host Mouse
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # MA121541
  • Size 50µg
Applications
Western Blot (Assay dependent)
Alternate names:
Somatomedin-C; Mechano growth factor
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA121541 375.00
Check your price
  • IGF1 Monoclonal Antibody for Western Blot and ELISA
  • Host Mouse
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # MA523752
  • Size 500µg
Applications
ELISA (0.1-0.4 µg/ml (Detection), 2-8 µg/ml (Capture)), Western Blot (1 µg/ml)
Alternate names:
Somatomedin-C; Mechano growth factor
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA523752 349.00
Check your price
  • IGF1 Monoclonal Antibody for IP
  • Host Mouse
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # MA121539
  • Size 50µg
Applications
Immunoprecipitation (Assay dependent)
Alternate names:
Somatomedin-C; Mechano growth factor
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA121539 375.00
Check your price
  • IGF1 Monoclonal Antibody for ELISA and RIA
  • Host Mouse
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # MA516806
  • Size 200ug
Applications
ELISA (Assay Dependent), Radioimmune Assays (Assay Dependent)
Alternate names:
Somatomedin-C; Mechano growth factor
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA516806 275.00
Check your price
  • Phospho-IR/IGF1R (Tyr1135, Tyr1136, Tyr1150, Tyr1151) Monoclonal Antibody for Western Blot
  • Host Rabbit
  • Target Species Bovine, Human, Mouse, Rat
  • Conjugate Unconjugated
  • Catalog # MA515148
  • Size 100µL
Applications
Western Blot (1:1000)
Alternate names:
Insulin-like growth factor I receptor
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA515148 309.00
Check your price
  • AKT3 Monoclonal Antibody for Western Blot and IP
  • Host Rabbit
  • Target Species Human, Mouse, Rat
  • Conjugate Unconjugated
  • Catalog # MA516309
  • Size 100µL
Applications
Immunoprecipitation (1:100), Western Blot (1:1000)
Alternate names:
RAC-PK-gamma; Protein kinase B gamma; Protein kinase Akt-3; STK-2
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA516309 275.00
Check your price
  • Phospho-IR/IGF1R (Tyr1162, Tyr1163) Monoclonal Antibody for Western Blot and ELISA
  • Host Rabbit
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # 700393
  • Size 100µg
Applications
ELISA (1-5 ug/ml), Western Blot (1-2 ug/ml)
Alternate names:
Insulin-like growth factor I receptor
Catalog # Size Price (AUD) Your Price (AUD) Qty
700393 309.00
Check your price
  • AKT1 Monoclonal Antibody for Western Blot and IHC (P)
  • Host Mouse
  • Target Species Human, Mouse, Rat
  • Conjugate Unconjugated
  • Catalog # AHO1112
  • Size 100µg
Applications
Immunohistochemistry (Paraffin) (1:10-1:100), Western Blot (Assay Dependent)
Alternate names:
RAC-PK-alpha; Protein kinase B alpha; Protein kinase B; Proto-oncogene c-Akt; Thymoma viral proto-oncogene; AKT1 kinase
Catalog # Size Price (AUD) Your Price (AUD) Qty
AHO1112 304.00
Check your price
  • AKT2 Monoclonal Antibody for Western Blot, IHC (P) and ELISA
  • Host Mouse
  • Target Species Human, Rat
  • Conjugate Unconjugated
  • Catalog # 393900
  • Size 100µg
Applications
ELISA (Assay Dependent), Immunohistochemistry (Paraffin) (1:50-1:500), Western Blot (Assay Dependent)
Alternate names:
RAC-PK-beta; Protein kinase B beta; Protein kinase Akt-2
Catalog # Size Price (AUD) Your Price (AUD) Qty
393900 302.00
Check your price
  • IGF1R beta Monoclonal Antibody for Western Blot and IHC
  • Host Mouse
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # MA515354
  • Size 100µL
Applications
Immunohistochemistry (1/200-1/1000), Western Blot (1/500-1/2000)
Alternate names:
Insulin-like growth factor I receptor
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA515354 349.00
Check your price
  • IGF1R beta Monoclonal Antibody for Western Blot and IHC
  • Host Mouse
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # MA515343
  • Size 100µL
Applications
Immunohistochemistry (1/200-1/1000), Western Blot (1/500-1/2000)
Alternate names:
Insulin-like growth factor I receptor
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA515343 349.00
Check your price
  • IGF1R alpha Monoclonal Antibody for Western Blot, IHC (P) and Flow
  • Host Mouse
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # MA181548
  • Size 200µg
Applications
Flow Cytometry (1/10), Immunohistochemistry (Paraffin) (1:10-1:40), Western Blot (Assay Dependent)
Alternate names:
Insulin-like growth factor I receptor
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA181548 375.00
Check your price
  • PRAS40 Monoclonal Antibody for Western Blot, IF, ICC, IHC (P) and IP
  • Host Mouse
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # AHO1031
  • Size 100µg
Applications
Immunocytochemistry (1:100-1:500), Immunofluorescence (1:100-1:500), Immunohistochemistry (Paraffin) (1:10-1:100), Immunoprecipitation (Assay Dependent), Western Blot (Assay Dependent)
Alternate names:
40 kDa proline-rich AKT substrate
Catalog # Size Price (AUD) Your Price (AUD) Qty
AHO1031 305.00
Check your price
  • IGF1R beta Monoclonal Antibody for Western Blot, IHC and ELISA
  • Host Mouse
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # 396700
  • Size 100µg
Applications
ELISA (Assay Dependent), Immunohistochemistry (Assay Dependent), Western Blot (2-3 ug/ml)
Alternate names:
Insulin-like growth factor I receptor
Catalog # Size Price (AUD) Your Price (AUD) Qty
396700 309.00
Check your price
  • IGF1R alpha Monoclonal Antibody for Western Blot and Neu
  • Host Mouse
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # MA121540
  • Size 50µg
Applications
Blocking Assay (Assay-dependent), Western Blot (Assay dependent)
Alternate names:
Insulin-like growth factor I receptor
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA121540 375.00
Check your price
  • IGFBP1 Monoclonal Antibody for Western Blot, ELISA and Neu
  • Host Mouse
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # MA124775
  • Size 100µg
Applications
ELISA (2 µg/ml), Neutralization (10-40 µg/ml), Western Blot (1-2 ug/ml)
Alternate names:
Placental protein 12
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA124775 309.00
Check your price
  • Phospho-AKT1 (Ser473) Monoclonal Antibody for Western Blot, IF, ICC, IHC (P), Flow and ELISA
  • Host Rabbit
  • Target Species Human, Mouse
  • Conjugate Unconjugated
  • Catalog # 700392
  • Size 100µg
Applications
ELISA (1-5 ug/ml), Flow Cytometry (1-3µg x 10^6 cells), Immunocytochemistry (1-2µg), Immunofluorescence (1-2µg), Immunohistochemistry (Paraffin) (0.5-2 ug/ml), Western Blot (1-3µg/ml)
Alternate names:
RAC-PK-alpha; Proto-oncogene c-Akt; Protein kinase B alpha; Protein kinase B; Thymoma viral proto-oncogene; AKT1 kinase
Catalog # Size Price (AUD) Your Price (AUD) Qty
700392 303.00
Check your price
  • Phospho-AKT1 (Ser473) Monoclonal Antibody for Western Blot, IF, ICC, Flow and ELISA
  • Host Rabbit
  • Target Species Human, Mouse
  • Conjugate Unconjugated
  • Catalog # 700256
  • Size 100µg
Applications
ELISA (1-5 ug/ml), Flow Cytometry (2-4µg x 10^6 cells), Immunocytochemistry (1-2µg), Immunofluorescence (1-2µg), Western Blot (0.5-1µg/ml)
Alternate names:
RAC-PK-alpha; Proto-oncogene c-Akt; Protein kinase B alpha; Protein kinase B; Thymoma viral proto-oncogene; AKT1 kinase
Catalog # Size Price (AUD) Your Price (AUD) Qty
700256 309.00
Check your price
  • Phospho-AKT1 (Thr308) Monoclonal Antibody for Western Blot, IF, ICC and Flow
  • Host Rabbit
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # 701052
  • Size 100µg
Applications
Flow Cytometry (1:100-1:500), Immunocytochemistry (1:100-1:1000), Immunofluorescence (1:100-1:1000), Western Blot (1:500-1:5000)
Alternate names:
RAC-PK-alpha; Proto-oncogene c-Akt; Protein kinase B alpha; Protein kinase B
Catalog # Size Price (AUD) Your Price (AUD) Qty
701052 304.00
Check your price
  • Phospho-FOXO4 (Thr28) Monoclonal Antibody for Western Blot
  • Host Rabbit
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # 701848
  • Size 100µg
Applications
Western Blot (0.5-1 µg/ml)
Alternate names:
Fork head domain transcription factor AFX1
Catalog # Size Price (AUD) Your Price (AUD) Qty
701848 325.00
Check your price
  • Phospho-p70 S6 Kinase (Thr421, Ser424) Monoclonal Antibody for Western Blot, Flow and ChIP
  • Host Rabbit
  • Target Species Human, Rat
  • Conjugate Unconjugated
  • Catalog # 701083
  • Size 100µg
Applications
ChIP assay (1 ul), Flow Cytometry (1:100-1:500), Western Blot (1:500-1:5000)
Alternate names:
p70 ribosomal S6 kinase alpha; Serine/threonine-protein kinase 14A; Ribosomal protein S6 kinase I; 70 kDa ribosomal protein S6 kinase 1
Catalog # Size Price (AUD) Your Price (AUD) Qty
701083 Contact Us ›
Check your price
  • Phospho-AS160 (Thr642) Monoclonal Antibody for Western Blot
  • Host Rabbit
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # 700565
  • Size 100µg
Applications
Western Blot (2-3 ug/ml)
Alternate names:
Akt substrate of 160 kDa
Catalog # Size Price (AUD) Your Price (AUD) Qty
700565 309.00
Check your price
  • Phospho-AKT1 (Thr308) Oligoclonal Antibody for Western Blot, IF and ICC
  • Host Rabbit
  • Target Species Human, Mouse
  • Conjugate Unconjugated
  • Catalog # 710122
  • Size 100µg
Applications
Immunocytochemistry (2-3 ug/ml), Immunofluorescence (2-3 ug/ml), Western Blot (0.5-2 ug/ml)
Alternate names:
RAC-PK-alpha; Proto-oncogene c-Akt; Protein kinase B alpha; Protein kinase B; Thymoma viral proto-oncogene; AKT1 kinase
Catalog # Size Price (AUD) Your Price (AUD) Qty
710122 309.00
Check your price
  • AKT1 Oligoclonal Antibody for Western Blot, IF, ICC, IHC (P) and Flow
  • Host Rabbit
  • Target Species Human, Mouse
  • Conjugate Unconjugated
  • Catalog # 710005
  • Size 100µg
Applications
Flow Cytometry (1:100-1:500), Immunocytochemistry (1:100-1:1000), Immunofluorescence (1:100-1:1000), Immunohistochemistry (Paraffin) (1:10-1:100), Western Blot (1:500-1:5000)
Alternate names:
RAC-PK-alpha; Proto-oncogene c-Akt; Protein kinase B alpha; Protein kinase B; Thymoma viral proto-oncogene; AKT1 kinase
Catalog # Size Price (AUD) Your Price (AUD) Qty
710005 309.00
Check your price
  • IGF1R beta Oligoclonal Antibody for Western Blot and ELISA
  • Host Rabbit
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # 710065
  • Size 100µg
Applications
ELISA (Assay-Dependent), Western Blot (2.5 ug/ml)
Alternate names:
Insulin-like growth factor I receptor
Catalog # Size Price (AUD) Your Price (AUD) Qty
710065 309.00
Check your price
  • INSR beta Oligoclonal Antibody for Western Blot
  • Host Rabbit
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # 710076
  • Size 100µg
Applications
Western Blot (2-5 ug/ml)
Alternate names:
CD220, HHF5, insulin receptor, INSR, CD221, IGF-I receptor, insulin-like growth factor I receptor, soluble IGF1R variant 1, soluble IGF1R variant 2
Catalog # Size Price (AUD) Your Price (AUD) Qty
710076 309.00
Check your price