1 - 30 of 30 products displayed
  • IGF1 Monoclonal Antibody for Western Blot, ELISA and Neu
  • Host Hamster
  • Target Species Mouse
  • Conjugate Unconjugated
  • Catalog # MA523772
  • Size 500 µg
Applications
ELISA (0.1-0.4 µg/ml (Detection), 2-8 µg/ml (Capture)), Neutralization (0.3-1.5 µg/ml), Western Blot (1 µg/ml)
Alternate names:
Somatomedin
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA523772 349.00
Check your price
  • IGF1 Polyclonal Antibody for Western Blot, IF, ICC and IHC (P)
  • Host Rabbit
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # PA527207
  • Size 100 µL
Applications
Immunocytochemistry (Assay-Dependent), Immunofluorescence (Assay-Dependent), Immunohistochemistry (Paraffin) (Assay-Dependent), Western Blot (1:500-1:3000)
Alternate names:
Somatomedin-C; Mechano growth factor
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA527207 349.00
Check your price
  • IGF1 Monoclonal Antibody for Western Blot and ELISA
  • Host Mouse
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # MA523752
  • Size 500 µg
Applications
ELISA (0.1-0.4 µg/ml (Detection), 2-8 µg/ml (Capture)), Western Blot (1 µg/ml)
Alternate names:
Somatomedin-C; Mechano growth factor
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA523752 349.00
Check your price
  • IGF1 Polyclonal Antibody for Western Blot and ELISA
  • Host Goat
  • Target Species Mouse
  • Conjugate Unconjugated
  • Catalog # PA132909
  • Size 100 µg
Applications
ELISA (Assay dependent), Western Blot (Assay dependent)
Alternate names:
Somatomedin
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA132909 600.00
Check your price
  • IGF1 Polyclonal Antibody, Biotin conjugate for Western Blot and ELISA
  • Host Rabbit
  • Target Species Human
  • Conjugate Biotin
  • Catalog # PA186912
  • Size 50 µg
Applications
ELISA (0.25ug/ml-1.0ug/ml), Western Blot (0.1ug/ml-0.2ug/ml)
Alternate names:
Somatomedin-C; Mechano growth factor
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA186912 499.00
Check your price
  • IGF1 Polyclonal Antibody for Western Blot, ELISA and FN
  • Host Rabbit
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # PA186913
  • Size 100 µg
Applications
ELISA (0.5-2.0 ug/ml), Functional Assay (0.67-1.0 µg/ml), Western Blot (0.1-0.2ug/ml)
Alternate names:
Somatomedin-C; Mechano growth factor
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA186913 550.00
Check your price
  • IGF1 (human) for Ctrl
  • Host E. Coli
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # RP8606
  • Size 100 µg
Applications
Control (Assay Dependent)
Alternate names:
Somatomedin-C; Mechano growth factor
Catalog # Size Price (AUD) Your Price (AUD) Qty
RP8606 314.00
Check your price
  • IGF1 for Ctrl
  • Host E. Coli
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # RP10910
  • Size 100 µg
Applications
Control (Assay Dependent)
Alternate names:
IGF-I, IGF-1, Somatomedin C
Catalog # Size Price (AUD) Your Price (AUD) Qty
RP10910 307.00
Check your price
  • IGF1 for Ctrl
  • Host E. coli
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # RP10931
  • Size 1000 µg
Applications
Control (Assay Dependent)
Alternate names:
IGF-I, IGF-1, Somatomedin C
Catalog # Size Price (AUD) Your Price (AUD) Qty
RP10931 429.00
Check your price
  • IGF1 (mouse) for Ctrl
  • Host E. coli
  • Target Species Mouse
  • Conjugate Unconjugated
  • Catalog # RP8603
  • Size 50 µg
Applications
Control (Assay Dependent)
Alternate names:
Somatomedin
Catalog # Size Price (AUD) Your Price (AUD) Qty
RP8603 269.00
Check your price
  • IGF1 (rat) for Ctrl
  • Host E. Coli
  • Target Species Rat
  • Conjugate Unconjugated
  • Catalog # RP8604
  • Size 50 µg
Applications
Control (Assay Dependent)
Alternate names:
Somatomedin
Catalog # Size Price (AUD) Your Price (AUD) Qty
RP8604 301.00
Check your price
  • IGF1 Monoclonal Antibody for Western Blot and ELISA
  • Host Mouse
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # MA1088
  • Size 200 ug
Applications
ELISA (1ug/ml), Western Blot (1:500-1:2000)
Alternate names:
Somatomedin-C; Mechano growth factor
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA1088 309.00
Check your price
  • IGF1 Polyclonal Antibody for Western Blot and ELISA
  • Host Rabbit
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # PA1130
  • Size 200 ug
Applications
ELISA (1ug/ml), Western Blot (1:500-1:2000)
Alternate names:
Somatomedin-C; Mechano growth factor
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA1130 309.00
Check your price
  • IGF1 Polyclonal Antibody for Western Blot, ELISA and Neu
  • Host Rabbit
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # PA130828
  • Size 100 µg
Applications
ELISA (2-3 µg/ml), Neutralization (0.5-4 µg/ml), Western Blot (0.5-4 µg/ml)
Alternate names:
Somatomedin-C; Mechano growth factor
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA130828 425.00
Check your price
  • IGF1 Monoclonal Antibody for Western Blot
  • Host Mouse
  • Target Species Human, Mouse, Rat
  • Conjugate Unconjugated
  • Catalog # MA512247
  • Size 500 µL
Applications
Western Blot (1-2 µg/ml)
Alternate names:
Somatomedin; Somatomedin-C; Mechano growth factor
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA512247 349.00
Check your price
  • IGF1 Receptor for Ctrl
  • Host E. Coli
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # RP39131
  • Size 10 µg
Applications
Control (Assay Dependent)
Alternate names:
Catalog # Size Price (AUD) Your Price (AUD) Qty
RP39131 375.00
Check your price
  • Phospho-IR/IGF1R (Tyr1162, Tyr1163) Monoclonal Antibody for Western Blot and ELISA
  • Host Rabbit
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # 700393
  • Size 100 µg
Applications
ELISA (1-5 ug/ml), Western Blot (1-2 ug/ml)
Alternate names:
Insulin-like growth factor I receptor
Catalog # Size Price (AUD) Your Price (AUD) Qty
700393 309.00
Check your price
  • IGF1R beta Monoclonal Antibody for Western Blot and IHC
  • Host Mouse
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # MA515354
  • Size 100 µL
Applications
Immunohistochemistry (1/200-1/1000), Western Blot (1/500-1/2000)
Alternate names:
Insulin-like growth factor I receptor
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA515354 349.00
Check your price
  • IGF1R beta Monoclonal Antibody for Western Blot and IHC
  • Host Mouse
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # MA515343
  • Size 100 µL
Applications
Immunohistochemistry (1/200-1/1000), Western Blot (1/500-1/2000)
Alternate names:
Insulin-like growth factor I receptor
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA515343 349.00
Check your price
  • INSR beta Oligoclonal Antibody for Western Blot
  • Host Rabbit
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # 710076
  • Size 100 µg
Applications
Western Blot (2-5 ug/ml)
Alternate names:
CD220, HHF5, insulin receptor, INSR, CD221, IGF-I receptor, insulin-like growth factor I receptor, soluble IGF1R variant 1, soluble IGF1R variant 2
Catalog # Size Price (AUD) Your Price (AUD) Qty
710076 309.00
Check your price
  • AKT3 Polyclonal Antibody for Western Blot, IF and ICC
  • Host Rabbit
  • Target Species Human, Mouse, Rat
  • Conjugate Unconjugated
  • Catalog # PA529695
  • Size 100 µL
Applications
Immunocytochemistry (1:100-1:1000), Immunofluorescence (1:100-1:1000), Western Blot (1:1000-1:10,000)
Alternate names:
RAC-PK-gamma; Protein kinase B gamma; Protein kinase Akt-3; STK-2
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA529695 349.00
Check your price
  • AKT3 Polyclonal Antibody for Western Blot, IF, ICC and IHC (P)
  • Host Rabbit
  • Target Species Human, Mouse
  • Conjugate Unconjugated
  • Catalog # PA529696
  • Size 100 µL
Applications
Immunocytochemistry (1:100-1:1000), Immunofluorescence (1:100-1:1000), Immunohistochemistry (Paraffin) (1:100-1:1000), Western Blot (1:1000-1:10,000)
Alternate names:
STK-2; RAC-PK-gamma; Protein kinase B gamma; Protein kinase Akt-3
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA529696 349.00
Check your price
  • IGF1R beta Oligoclonal Antibody for Western Blot and ELISA
  • Host Rabbit
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # 710065
  • Size 100 µg
Applications
ELISA (Assay-Dependent), Western Blot (2.5 ug/ml)
Alternate names:
Insulin-like growth factor I receptor
Catalog # Size Price (AUD) Your Price (AUD) Qty
710065 309.00
Check your price
  • AKT1 Monoclonal Antibody for Western Blot and IHC (P)
  • Host Mouse
  • Target Species Human, Mouse, Rat
  • Conjugate Unconjugated
  • Catalog # AHO1112
  • Size 100 µg
Applications
Immunohistochemistry (Paraffin) (1:10-1:100), Western Blot (Assay Dependent)
Alternate names:
RAC-PK-alpha; Protein kinase B alpha; Protein kinase B; Proto-oncogene c-Akt; Thymoma viral proto-oncogene; AKT1 kinase
Catalog # Size Price (AUD) Your Price (AUD) Qty
AHO1112 304.00
Check your price
  • PRAS40 Monoclonal Antibody for Western Blot, IF, ICC, IHC (P) and IP
  • Host Mouse
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # AHO1031
  • Size 100 µg
Applications
Immunocytochemistry (1:100-1:500), Immunofluorescence (1:100-1:500), Immunohistochemistry (Paraffin) (1:10-1:100), Immunoprecipitation (Assay Dependent), Western Blot (Assay Dependent)
Alternate names:
40 kDa proline-rich AKT substrate
Catalog # Size Price (AUD) Your Price (AUD) Qty
AHO1031 305.00
Check your price
  • IGFBP1 Monoclonal Antibody for Western Blot, ELISA and Neu
  • Host Mouse
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # MA124775
  • Size 100 µg
Applications
ELISA (2 µg/ml), Neutralization (10-40 µg/ml), Western Blot (1-2 ug/ml)
Alternate names:
Placental protein 12
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA124775 309.00
Check your price
  • Phospho-AKT1 (Thr308) Oligoclonal Antibody for Western Blot, IF and ICC
  • Host Rabbit
  • Target Species Human, Mouse
  • Conjugate Unconjugated
  • Catalog # 710122
  • Size 100 µg
Applications
Immunocytochemistry (2-3 ug/ml), Immunofluorescence (2-3 ug/ml), Western Blot (0.5-2 ug/ml)
Alternate names:
RAC-PK-alpha; Proto-oncogene c-Akt; Protein kinase B alpha; Protein kinase B; Thymoma viral proto-oncogene; AKT1 kinase
Catalog # Size Price (AUD) Your Price (AUD) Qty
710122 309.00
Check your price
  • AKT1 Oligoclonal Antibody for Western Blot, IF, ICC, IHC (P) and Flow
  • Host Rabbit
  • Target Species Human, Mouse
  • Conjugate Unconjugated
  • Catalog # 710005
  • Size 100 µg
Applications
Flow Cytometry (1:100-1:500), Immunocytochemistry (1:100-1:1000), Immunofluorescence (1:100-1:1000), Immunohistochemistry (Paraffin) (1:10-1:100), Western Blot (1:500-1:5000)
Alternate names:
RAC-PK-alpha; Proto-oncogene c-Akt; Protein kinase B alpha; Protein kinase B; Thymoma viral proto-oncogene; AKT1 kinase
Catalog # Size Price (AUD) Your Price (AUD) Qty
710005 309.00
Check your price
  • Phospho-AKT1 (Ser473) Monoclonal Antibody for Western Blot, IF, ICC, IHC (P), Flow and ELISA
  • Host Rabbit
  • Target Species Human, Mouse
  • Conjugate Unconjugated
  • Catalog # 700392
  • Size 100 µg
Applications
ELISA (1-5 ug/ml), Flow Cytometry (1-3µg x 10^6 cells), Immunocytochemistry (1-2µg), Immunofluorescence (1-2µg), Immunohistochemistry (Paraffin) (0.5-2 ug/ml), Western Blot (1-3µg/ml)
Alternate names:
RAC-PK-alpha; Proto-oncogene c-Akt; Protein kinase B alpha; Protein kinase B; Thymoma viral proto-oncogene; AKT1 kinase
Catalog # Size Price (AUD) Your Price (AUD) Qty
700392 303.00
Check your price
  • Phospho-AKT1 (Ser473) Monoclonal Antibody for Western Blot, IF, ICC, Flow and ELISA
  • Host Rabbit
  • Target Species Human, Mouse
  • Conjugate Unconjugated
  • Catalog # 700256
  • Size 100 µg
Applications
ELISA (1-5 ug/ml), Flow Cytometry (2-4µg x 10^6 cells), Immunocytochemistry (1-2µg), Immunofluorescence (1-2µg), Western Blot (0.5-1µg/ml)
Alternate names:
RAC-PK-alpha; Proto-oncogene c-Akt; Protein kinase B alpha; Protein kinase B; Thymoma viral proto-oncogene; AKT1 kinase
Catalog # Size Price (AUD) Your Price (AUD) Qty
700256 309.00
Check your price