1 - 30 of 30 products displayed
  • IGF1R alpha Polyclonal Antibody for Western Blot, IF, ICC, IHC (P) and Flow
  • Host Rabbit
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # PA524610
  • Size 400µL
Applications
Flow Cytometry (1:10-1:50), Immunocytochemistry (1:10-1:50), Immunofluorescence (1:10-1:50), Immunohistochemistry (1:50-1:100), Western Blot (1:1000)
Alternate names:
Insulin-like growth factor I receptor
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA524610 349.00
Check your price
  • IGF1R alpha Polyclonal Antibody for Western Blot and IHC
  • Host Rabbit
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # PA514677
  • Size 400µL
Applications
Immunohistochemistry (1:50-1:100), Western Blot (1:1000)
Alternate names:
Insulin-like growth factor I receptor
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA514677 349.00
Check your price
  • IGF1R beta Monoclonal Antibody for Western Blot and IHC
  • Host Mouse
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # MA515354
  • Size 100µL
Applications
Immunohistochemistry (1/200-1/1000), Western Blot (1/500-1/2000)
Alternate names:
Insulin-like growth factor I receptor
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA515354 349.00
Check your price
  • IGF1R beta Monoclonal Antibody for Western Blot and IHC
  • Host Mouse
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # MA515343
  • Size 100µL
Applications
Immunohistochemistry (1/200-1/1000), Western Blot (1/500-1/2000)
Alternate names:
Insulin-like growth factor I receptor
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA515343 349.00
Check your price
  • IGF1R beta Oligoclonal Antibody for Western Blot and ELISA
  • Host Rabbit
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # 710065
  • Size 100µg
Applications
ELISA (Assay-Dependent), Western Blot (2.5 ug/ml)
Alternate names:
Insulin-like growth factor I receptor
Catalog # Size Price (AUD) Your Price (AUD) Qty
710065 309.00
Check your price
  • Phospho-IGF1R beta (Tyr1135, Tyr1136) Monoclonal Antibody for Western Blot
  • Host Rabbit
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # 701067
  • Size 100µg
Applications
Western Blot (1-3 ug/ml)
Alternate names:
Insulin-like growth factor I receptor
Catalog # Size Price (AUD) Your Price (AUD) Qty
701067 309.00
Check your price
  • Phospho-IGF1R beta (Tyr1346) Polyclonal Antibody for IHC (P)
  • Host Rabbit
  • Target Species Human, Mouse, Rat
  • Conjugate Unconjugated
  • Catalog # PA537604
  • Size 100µl
Applications
Immunohistochemistry (Paraffin) (1:50-1:100)
Alternate names:
Insulin-like growth factor I receptor
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA537604 309.00
Check your price
  • Phospho-IGF1R beta (Tyr1158, Tyr1162, Tyr1163) Polyclonal Antibody for Western Blot
  • Host Rabbit
  • Target Species Human, Mouse
  • Conjugate Unconjugated
  • Catalog # PA126725
  • Size 50µL
Applications
Western Blot (1:1000)
Alternate names:
Insulin-like growth factor I receptor
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA126725 349.00
Check your price
  • Phospho-IR/IGF1R (Tyr1162, Tyr1163) Polyclonal Antibody for Western Blot, IF, ICC, IHC and Flow
  • Host Rabbit
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # 44804G
  • Size 10blots
Applications
Flow Cytometry (1:100-1:500), Immunocytochemistry (1:100-1:500), Immunofluorescence (1:100-1:500), Immunohistochemistry (Assay Dependent), Western Blot (Assay Dependent)
Alternate names:
Insulin-like growth factor I receptor
Catalog # Size Price (AUD) Your Price (AUD) Qty
44804G 304.00
Check your price
  • IGF1R beta Monoclonal Antibody for Western Blot, IHC and ELISA
  • Host Mouse
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # 396700
  • Size 100µg
Applications
ELISA (Assay Dependent), Immunohistochemistry (Assay Dependent), Western Blot (2-3 ug/ml)
Alternate names:
Insulin-like growth factor I receptor
Catalog # Size Price (AUD) Your Price (AUD) Qty
396700 309.00
Check your price
  • Phospho-IR/IGF1R (Tyr1158) Polyclonal Antibody for Western Blot, IF, ICC and IHC
  • Host Rabbit
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # 44802G
  • Size 10blots
Applications
Immunocytochemistry (1:100-1:500), Immunofluorescence (1:100-1:500), Immunohistochemistry (Assay Dependent), Western Blot (Assay Dependent)
Alternate names:
Insulin-like growth factor I receptor
Catalog # Size Price (AUD) Your Price (AUD) Qty
44802G 304.00
Check your price
  • Phospho-IR/IGF1R (Tyr1158, Tyr1162, Tyr1163) Polyclonal Antibody for Western Blot
  • Host Rabbit
  • Target Species Human, Mouse
  • Conjugate Unconjugated
  • Catalog # 44806G
  • Size 10blots
Applications
Western Blot (Assay Dependent)
Alternate names:
Insulin-like growth factor I receptor
Catalog # Size Price (AUD) Your Price (AUD) Qty
44806G 304.00
Check your price
  • Phospho-IGF1R beta (Tyr1162, Tyr1163) Polyclonal Antibody for Western Blot
  • Host Rabbit
  • Target Species Human, Mouse, Rat
  • Conjugate Unconjugated
  • Catalog # PA126724
  • Size 50µL
Applications
Western Blot (1:1000)
Alternate names:
Insulin-like growth factor I receptor
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA126724 349.00
Check your price
  • Phospho-IR/IGF1R (Tyr1135, Tyr1136, Tyr1150, Tyr1151) Monoclonal Antibody for Western Blot
  • Host Rabbit
  • Target Species Bovine, Human, Mouse, Rat
  • Conjugate Unconjugated
  • Catalog # MA515148
  • Size 100µL
Applications
Western Blot (1:1000)
Alternate names:
Insulin-like growth factor I receptor
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA515148 309.00
Check your price
  • Phospho-IGF1R beta (Tyr1165, Tyr1166) Polyclonal Antibody for Western Blot, IF and ICC
  • Host Rabbit
  • Target Species Human, Mouse, Rat
  • Conjugate Unconjugated
  • Catalog # PA537602
  • Size 100µl
Applications
Immunocytochemistry (1:100-1:200), Immunofluorescence (1:100-1:200), Western Blot (1:500-1:1000)
Alternate names:
Insulin-like growth factor I receptor
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA537602 309.00
Check your price
  • Phospho-IGF1R beta (Tyr1161) Polyclonal Antibody for Western Blot, IF, ICC and IHC (P)
  • Host Rabbit
  • Target Species Human, Mouse, Rat
  • Conjugate Unconjugated
  • Catalog # PA537601
  • Size 100µl
Applications
Immunocytochemistry (1:100-1:200), Immunofluorescence (1:100-1:200), Immunohistochemistry (Paraffin) (1:50-1:100), Western Blot (1:500-1:1000)
Alternate names:
Insulin-like growth factor I receptor
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA537601 309.00
Check your price
  • Phospho-IGF1R beta (Tyr1280) Polyclonal Antibody for Western Blot
  • Host Rabbit
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # PA537603
  • Size 100µl
Applications
Western Blot (1:500-1:1000)
Alternate names:
Insulin-like growth factor I receptor
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA537603 309.00
Check your price
  • IGF1R alpha Monoclonal Antibody for Western Blot, IHC (P) and Flow
  • Host Mouse
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # MA181548
  • Size 200µg
Applications
Flow Cytometry (1/10), Immunohistochemistry (Paraffin) (1:10-1:40), Western Blot (Assay Dependent)
Alternate names:
Insulin-like growth factor I receptor
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA181548 375.00
Check your price
  • Phospho-IR/IGF1R (Tyr1162, Tyr1163) Monoclonal Antibody for Western Blot and ELISA
  • Host Rabbit
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # 700393
  • Size 100µg
Applications
ELISA (1-5 ug/ml), Western Blot (1-2 ug/ml)
Alternate names:
Insulin-like growth factor I receptor
Catalog # Size Price (AUD) Your Price (AUD) Qty
700393 309.00
Check your price
  • IGF1R alpha Monoclonal Antibody for IF, ICC, IP and ELISA
  • Host Mouse
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # AHR0321
  • Size 500µl
Applications
ELISA (Assay Dependent), Immunocytochemistry (1:10-1:100), Immunofluorescence (Assay-Dependent), Immunoprecipitation (2µg per mg lysate)
Alternate names:
Insulin-like growth factor I receptor
Catalog # Size Price (AUD) Your Price (AUD) Qty
AHR0321 304.00
Check your price
  • IGF1R alpha Monoclonal Antibody, Biotin conjugate for IF, ICC, IHC (P), IP and ELISA
  • Host Mouse
  • Target Species Human
  • Conjugate Biotin
  • Catalog # MA513799
  • Size 500µL
Applications
ELISA (Assay Dependent), Immunocytochemistry (1:10-1:100), Immunofluorescence (1:10-1:100), Immunohistochemistry (Paraffin) (Assay Dependent), Immunoprecipitation (2 µg/ml)
Alternate names:
Insulin-like growth factor I receptor
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA513799 399.00
Check your price
  • IGF1R alpha Monoclonal Antibody for Western Blot, IF, ICC, IP and IA
  • Host Mouse
  • Target Species Human, Mouse
  • Conjugate Unconjugated
  • Catalog # MA513817
  • Size 500µL
Applications
Immunocytochemistry (1:10-1:100), Immunofluorescence (1:10-1:100), Immunoprecipitation (2 µg/ml), Inhibition Assays (1.5 µg/mL), Western Blot (1:10-1:200)
Alternate names:
Insulin-like growth factor I receptor
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA513817 349.00
Check your price
  • IGF1R beta Monoclonal Antibody for Western Blot
  • Host Mouse
  • Target Species Human, Mouse, Rat
  • Conjugate Unconjugated
  • Catalog # AHO1292
  • Size 100µg
Applications
Western Blot (Assay Dependent)
Alternate names:
Insulin-like growth factor I receptor
Catalog # Size Price (AUD) Your Price (AUD) Qty
AHO1292 304.00
Check your price
  • Phospho-IGF1R beta(Tyr1316) Polyclonal Antibody for Western Blot and IP
  • Host Rabbit
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # A16700
  • Size 100µL
Applications
Immunoprecipitation (Assay Dependent), Western Blot (Assay Dependent)
Alternate names:
Insulin-like growth factor I receptor
Catalog # Size Price (AUD) Your Price (AUD) Qty
A16700 309.00
Check your price
  • Phospho-IGF1R beta (Tyr980) Monoclonal Antibody for Western Blot
  • Host Rabbit
  • Target Species Human, Mouse, Rat
  • Conjugate Unconjugated
  • Catalog # MA514859
  • Size 100µL
Applications
Western Blot (1:1000)
Alternate names:
Insulin-like growth factor I receptor
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA514859 309.00
Check your price
  • IGF1R alpha Monoclonal Antibody, PE conjugate for Flow
  • Host Mouse
  • Target Species Human
  • Conjugate PE
  • Catalog # MA516465
  • Size 100TESTS
Applications
Flow Cytometry (Neat)
Alternate names:
Insulin-like growth factor I receptor
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA516465 349.00
Check your price
  • IGF1R alpha Monoclonal Antibody for Western Blot and Neu
  • Host Mouse
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # MA121540
  • Size 50µg
Applications
Blocking Assay (Assay-dependent), Western Blot (Assay dependent)
Alternate names:
Insulin-like growth factor I receptor
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA121540 375.00
Check your price
  • IGF1R alpha Monoclonal Antibody for IP and IA
  • Host Mouse
  • Target Species Human, Pig, Rabbit
  • Conjugate Unconjugated
  • Catalog # MA513807
  • Size 500µL
Applications
Immunoprecipitation (2 µg/ml), Inhibition Assays (Assay Dependent)
Alternate names:
Insulin-like growth factor I receptor
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA513807 349.00
Check your price
  • IGF1R alpha Monoclonal Antibody for IP and IA
  • Host Mouse
  • Target Species Human, Rabbit
  • Conjugate Unconjugated
  • Catalog # MA513812
  • Size 500µL
Applications
Immunoprecipitation (Assay Dependent), Inhibition Assays (1.5 µg/mL)
Alternate names:
Insulin-like growth factor I receptor
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA513812 349.00
Check your price
  • IGF1R beta Monoclonal Antibody for Western Blot
  • Host Mouse
  • Target Species Human, Rat
  • Conjugate Unconjugated
  • Catalog # MA513822
  • Size 500µL
Applications
Western Blot (1-2 µg/ml)
Alternate names:
Insulin-like growth factor I receptor
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA513822 349.00
Check your price