1 - 21 of 21 products displayed
  • IGFBP1 Monoclonal Antibody for Western Blot, ELISA and Neu
  • Host Mouse
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # MA523727
  • Size 500µg
Applications
ELISA (0.1-0.4 µg/ml (Detection), 2-8 µg/ml (Capture)), Neutralization (10-40 µg/ml), Western Blot (1 µg/ml)
Alternate names:
Placental protein 12
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA523727 375.00
Check your price
  • IGFBP1 Polyclonal Antibody for Western Blot and IHC (F)
  • Host Goat
  • Target Species Mouse
  • Conjugate Unconjugated
  • Catalog # PA547102
  • Size 100µg
Applications
Immunohistochemistry (Frozen) (5-15 µg/ml), Western Blot (0.1 µg/ml)
Alternate names:
AFBP, IBP1, IGF-BP25, PP12, hIGFBP-1
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA547102 349.00
Check your price
  • IGFBP1 Monoclonal Antibody for Western Blot, ELISA and Neu
  • Host Mouse
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # MA124775
  • Size 100µg
Applications
ELISA (2 µg/ml), Neutralization (10-40 µg/ml), Western Blot (1-2 ug/ml)
Alternate names:
Placental protein 12
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA124775 309.00
Check your price
  • IGFBP5 Polyclonal Antibody for Western Blot and IHC (P)
  • Host Rabbit
  • Target Species Human, Mouse, Rat
  • Conjugate Unconjugated
  • Catalog # PA537369
  • Size 100µl
Applications
Immunohistochemistry (Paraffin) (1:50-1:200), Western Blot (1:500-1:2000)
Alternate names:
IBP 5, IBP-5, IBP5, IBP5, IGF binding protein 5, IGF BP5, IGF-binding protein 5, IGFBP 5, IGFBP-5, IGFBP5, Insulin Like Growth Factor Binding Protein 5, Insulin-like growth factor-binding protein 5
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA537369 399.00
Check your price
  • Phospho-FOXO1/FOXO3/FOXO4 (Thr24, Thr32) Polyclonal Antibody for IHC (P)
  • Host Rabbit
  • Target Species Human, Mouse, Rat
  • Conjugate Unconjugated
  • Catalog # PA536737
  • Size 100µl
Applications
Immunohistochemistry (Paraffin) (1:50-1:200)
Alternate names:
Fork head domain transcription factor AFX1; Forkhead in rhabdomyosarcoma; Forkhead box protein O1A; Afxh; Forkhead in rhabdomyosarcoma-like 1; AF6q21 protein
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA536737 349.00
Check your price
  • Phospho-FOXO pan pThr24 / pThr32 Polyclonal Antibody for Western Blot
  • Host Rabbit
  • Target Species Rat, Human, Mouse
  • Conjugate
  • Catalog # PA535576
  • Size 100µl
Applications
Western Blot (1:500-1:2000)
Alternate names:
Fork head domain transcription factor AFX1; Forkhead in rhabdomyosarcoma; Forkhead box protein O1A; Afxh; Forkhead in rhabdomyosarcoma-like 1; AF6q21 protein
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA535576 Not Orderable Online
Check your price
  • FOXO1/FOXO3/FOXO4 Polyclonal Antibody for Western Blot and IHC (P)
  • Host Rabbit
  • Target Species Human, Mouse, Rat
  • Conjugate Unconjugated
  • Catalog # PA536084
  • Size 100µl
Applications
Immunohistochemistry (Paraffin) (1:50-1:200), Western Blot (1:500-1:1000)
Alternate names:
Fork head domain transcription factor AFX1; Forkhead in rhabdomyosarcoma; Forkhead box protein O1A; Afxh; Forkhead in rhabdomyosarcoma-like 1; AF6q21 protein
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA536084 309.00
Check your price
  • Phospho-FOXO4 (Thr451) Polyclonal Antibody for Western Blot
  • Host Rabbit
  • Target Species Human, Mouse
  • Conjugate Unconjugated
  • Catalog # PA536804
  • Size 100µl
Applications
Western Blot (1:500-1:1000)
Alternate names:
Fork head domain transcription factor AFX1; Afxh
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA536804 349.00
Check your price
  • Phospho-FOXO4 (Ser197) Polyclonal Antibody for Western Blot and IHC (P)
  • Host Rabbit
  • Target Species Human, Mouse, Rat
  • Conjugate Unconjugated
  • Catalog # PA536817
  • Size 100µl
Applications
Immunohistochemistry (Paraffin) (1:50-1:200), Western Blot (1:500-1:1000)
Alternate names:
Fork head domain transcription factor AFX1; Afxh
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA536817 349.00
Check your price
  • Phospho-FOXO4 (Thr28) Oligoclonal Antibody for Western Blot, IF, ICC and IHC (P)
  • Host Rabbit
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # 710880
  • Size 100µg
Applications
Immunocytochemistry (1 µg/ml), Immunofluorescence (1 µg/ml), Western Blot (1-3 µg/ml)
Alternate names:
Fork head domain transcription factor AFX1
Catalog # Size Price (AUD) Your Price (AUD) Qty
710880 325.00
Check your price
  • Phospho-FOXO4 (Thr28) Monoclonal Antibody for Western Blot
  • Host Rabbit
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # 701848
  • Size 100µg
Applications
Western Blot (0.5-1 µg/ml)
Alternate names:
Fork head domain transcription factor AFX1
Catalog # Size Price (AUD) Your Price (AUD) Qty
701848 325.00
Check your price
  • FOXO4 Polyclonal Antibody for Western Blot, IF, ICC and ChIP
  • Host Rabbit
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # 720154
  • Size 100µg
Applications
ChIP assay (3 µg/ml), Immunocytochemistry (2 µg/ml), Immunofluorescence (2 µg/ml), Western Blot (2-3 µg/ml)
Alternate names:
Fork head domain transcription factor AFX1
Catalog # Size Price (AUD) Your Price (AUD) Qty
720154 309.00
Check your price
  • IGFBP5 Polyclonal Antibody for Western Blot, IHC (F) and ELISA
  • Host Goat
  • Target Species Mouse
  • Conjugate Unconjugated
  • Catalog # PA547775
  • Size 100µg
Applications
ELISA (0.1-0.4 µg/ml (Detection), 0.2-0.8 µg/ml (Capture)), Immunohistochemistry (Frozen) (5-15 µg/ml), Western Blot (0.1 µg/ml)
Alternate names:
IBP 5, IBP-5, IBP5, IBP5, IGF binding protein 5, IGF BP5, IGF-binding protein 5, IGFBP 5, IGFBP-5, IGFBP5, Insulin Like Growth Factor Binding Protein 5, Insulin-like growth factor-binding protein 5
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA547775 349.00
Check your price
  • Phospho-FOXO1/FOXO3/FOXO4 (Thr24, Thr32) Polyclonal Antibody for Western Blot
  • Host Rabbit
  • Target Species Human, Mouse, Rat
  • Conjugate Unconjugated
  • Catalog # PA537817
  • Size 100µl
Applications
Western Blot (1:500-1:1000)
Alternate names:
Fork head domain transcription factor AFX1; Forkhead in rhabdomyosarcoma; Forkhead box protein O1A; Afxh; Forkhead in rhabdomyosarcoma-like 1; AF6q21 protein
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA537817 309.00
Check your price
  • FOXO1/FOXO3/FOXO4 Polyclonal Antibody for IHC (P)
  • Host Rabbit
  • Target Species Human, Mouse
  • Conjugate Unconjugated
  • Catalog # PA538274
  • Size 100µg
Applications
Immunohistochemistry (Paraffin) (1:50-1:100)
Alternate names:
Fork head domain transcription factor AFX1; Forkhead in rhabdomyosarcoma; Forkhead box protein O1A; Afxh; Forkhead in rhabdomyosarcoma-like 1; AF6q21 protein
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA538274 309.00
Check your price
  • Phospho-FOXO1 (Ser329) Polyclonal Antibody for Western Blot and IHC (P)
  • Host Rabbit
  • Target Species Human, Mouse, Rat
  • Conjugate Unconjugated
  • Catalog # PA538275
  • Size 100µg
Applications
Immunohistochemistry (Paraffin) (1:50-1:100), Western Blot (1:500-1:1000)
Alternate names:
Forkhead in rhabdomyosarcoma; Forkhead box protein O1A
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA538275 309.00
Check your price
  • Phospho-FOXO1 (Ser319) Polyclonal Antibody for Western Blot, IF, ICC and IHC (P)
  • Host Rabbit
  • Target Species Human, Mouse, Rat
  • Conjugate Unconjugated
  • Catalog # PA537577
  • Size 100µl
Applications
Immunocytochemistry (1:100-1:200), Immunofluorescence (1:100-1:200), Immunohistochemistry (Paraffin) (1:50-1:100), Western Blot (1:500-1:1000)
Alternate names:
Forkhead in rhabdomyosarcoma; Forkhead box protein O1A
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA537577 309.00
Check your price
  • Phospho-FOXO1 (Ser256) Polyclonal Antibody for Western Blot and IHC (P)
  • Host Rabbit
  • Target Species Human, Mouse, Rat
  • Conjugate Unconjugated
  • Catalog # PA538132
  • Size 100µg
Applications
Immunohistochemistry (Paraffin) (1:50-1:100), Western Blot (1:500-1:1000)
Alternate names:
Forkhead in rhabdomyosarcoma; Forkhead box protein O1A
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA538132 309.00
Check your price
  • Phospho-FOXO4 (Ser262) Polyclonal Antibody for Western Blot and IHC (P)
  • Host Rabbit
  • Target Species Human, Mouse
  • Conjugate Unconjugated
  • Catalog # PA539717
  • Size 100µg
Applications
Immunohistochemistry (Paraffin) (1:50-1:100), Western Blot (1:500-1:1000)
Alternate names:
Fork head domain transcription factor AFX1; Afxh
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA539717 309.00
Check your price
  • Phospho-FOXO4 (Ser197) Polyclonal Antibody for Western Blot, IF, ICC and IHC (P)
  • Host Rabbit
  • Target Species Human, Mouse, Rat
  • Conjugate Unconjugated
  • Catalog # PA537648
  • Size 100µl
Applications
Immunocytochemistry (1:100-1:200), Immunofluorescence (1:100-1:200), Immunohistochemistry (Paraffin) (1:50-1:100), Western Blot (1:500-1:1000)
Alternate names:
Fork head domain transcription factor AFX1; Afxh
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA537648 309.00
Check your price
  • IGFBP5 Monoclonal Antibody for Western Blot and ELISA
  • Host Mouse
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # MA523872
  • Size 100µg
Applications
ELISA (0.1-0.4 µg/ml (Detection), 2-8 µg/ml (Capture)), Western Blot (1 µg/ml)
Alternate names:
IBP 5, IBP-5, IBP5, IBP5, IGF binding protein 5, IGF BP5, IGF-binding protein 5, IGFBP 5, IGFBP-5, IGFBP5, Insulin Like Growth Factor Binding Protein 5, Insulin-like growth factor-binding protein 5
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA523872 275.00
Check your price