1 - 30 of 30 products displayed
  • Rat anti-Mouse IgG1 Secondary Antibody for ELISA
  • Host Rat
  • Target Species Mouse
  • Conjugate Unconjugated
  • Catalog # MA516782
  • Size 1mg
Applications
ELISA (5 ug/ml as captive antibo)
Alternate names:
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA516782 428.00
Check your price
  • Mouse anti-Rat IgG1 Secondary Antibody, Biotin conjugate for ELISA
  • Host Mouse
  • Target Species Rat
  • Conjugate Biotin
  • Catalog # SA125262
  • Size 125µg
Applications
ELISA (0.5 µg/ml)
Alternate names:
Catalog # Size Price (AUD) Your Price (AUD) Qty
SA125262 234.00
Check your price
  • Goat anti-Mouse IgG1 Secondary Antibody, HRP conjugate for Western Blot, IHC (F), IHC (P) and ELISA
  • Host Goat
  • Target Species Mouse
  • Conjugate HRP
  • Catalog # PA174421
  • Size 1mL
Applications
ELISA (1:4000-1:8000), Immunohistochemistry (Frozen) (Assay Dependent), Immunohistochemistry (Paraffin) (Assay Dependent), Western Blot (Assay Dependent)
Alternate names:
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA174421 299.00
Check your price
  • Sheep anti-Bovine IgG1 Secondary Antibody, HRP conjugate for ELISA
  • Host Sheep
  • Target Species Bovine
  • Conjugate HRP
  • Catalog # PA184658
  • Size 1mg
Applications
ELISA (1/10,000-1/100,000)
Alternate names:
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA184658 360.00
Check your price
  • Sheep anti-Human IgG1 Secondary Antibody for Western Blot and ELISA
  • Host Sheep
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # PA533139
  • Size 500ug
Applications
ELISA (1:4000-1:8000), Western Blot (1:4000-1:8000)
Alternate names:
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA533139 682.00
Check your price
  • Mouse anti-Human IgG1 Secondary Antibody for IHC (F) and ELISA
  • Host Mouse
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # MA134581
  • Size 1mg
Applications
ELISA (Assay Dependent), Immunohistochemistry (Frozen) (Assay Dependent)
Alternate names:
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA134581 472.00
Check your price
  • Mouse anti-Rat IgG1 Secondary Antibody, Biotin conjugate for Flow
  • Host Mouse
  • Target Species Rat
  • Conjugate Biotin
  • Catalog # MA133298
  • Size 100tests
Applications
Flow Cytometry (10 µl for 10^6 cells in 100 µl)
Alternate names:
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA133298 304.00
Check your price
  • Mouse anti-Rat IgG1 Secondary Antibody for IHC (F) and Flow
  • Host Mouse
  • Target Species Rat
  • Conjugate Unconjugated
  • Catalog # MA133304
  • Size 250µg
Applications
Flow Cytometry (10 µl for 10^6 cells in 100 µl), Immunohistochemistry (Frozen) (Assay dependent)
Alternate names:
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA133304 550.00
Check your price
  • Mouse anti-Rat IgG1 Secondary Antibody for IHC (F) and Flow
  • Host Mouse
  • Target Species Rat
  • Conjugate Unconjugated
  • Catalog # MA133300
  • Size 100tests
Applications
Flow Cytometry (10 µl for 10^6 cells in 100 µl), Immunohistochemistry (Frozen) (Assay dependent)
Alternate names:
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA133300 309.00
Check your price
  • Rat anti-Mouse IgG1 Secondary Antibody, Biotin conjugate for IF
  • Host Rat
  • Target Species Mouse
  • Conjugate Biotin
  • Catalog # RMG115
  • Size 500µL
Applications
Immunofluorescence (0.5-2 µg/mL)
Alternate names:
Catalog # Size Price (AUD) Your Price (AUD) Qty
RMG115 154.00
Check your price
  • Rat anti-Mouse IgG1 Secondary Antibody, FITC conjugate for IF and Flow
  • Host Rat
  • Target Species Mouse
  • Conjugate FITC
  • Catalog # RMG101
  • Size 500µL
Applications
Flow Cytometry (0.5-2 µg/mL), Immunofluorescence (0.5-2 µg/mL)
Alternate names:
Catalog # Size Price (AUD) Your Price (AUD) Qty
RMG101 123.00
Check your price
  • Mouse anti-Bovine IgG1 Secondary Antibody, HRP conjugate for ELISA
  • Host Mouse
  • Target Species Bovine
  • Conjugate HRP
  • Catalog # MA516736
  • Size 250ug
Applications
ELISA (1:100-1:1000)
Alternate names:
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA516736 399.00
Check your price
  • Rat anti-Mouse IgG1 Secondary Antibody, HRP conjugate for IHC (F) and ELISA
  • Host Rat
  • Target Species Mouse
  • Conjugate HRP
  • Catalog # SA135640
  • Size 1mg
Applications
ELISA (Assay dependent.), Immunohistochemistry (Frozen) (Assay dependent.)
Alternate names:
Catalog # Size Price (AUD) Your Price (AUD) Qty
SA135640 1,612.00
Check your price
  • Rabbit anti-Mouse IgG1 Secondary Antibody, HRP conjugate for Western Blot, IHC (F) and ELISA
  • Host Rabbit
  • Target Species Mouse
  • Conjugate HRP
  • Catalog # PA186329
  • Size 1mg
Applications
ELISA (1:20,000-1:100,000), Immunohistochemistry (Frozen) (Assay Dependent), Western Blot (1:2000-1:10,000)
Alternate names:
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA186329 550.00
Check your price
  • Rat anti-Mouse IgG1 (Heavy chain) Secondary Antibody, HRP conjugate for Western Blot and ELISA
  • Host Rat
  • Target Species Mouse
  • Conjugate HRP
  • Catalog # MA516786
  • Size 500ug
Applications
ELISA (500 ng/ml)
Alternate names:
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA516786 650.00
Check your price
  • Rat anti-Mouse IgG1 (Heavy chain) Secondary Antibody, FITC conjugate for IF, ICC and Flow
  • Host Rat
  • Target Species Mouse
  • Conjugate FITC
  • Catalog # MA516785
  • Size 500ug
Applications
Flow Cytometry (5 ug/ml)
Alternate names:
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA516785 399.00
Check your price
  • Mouse anti-Rat IgG2a Secondary Antibody, FITC conjugate for Flow
  • Host Mouse
  • Target Species Rat
  • Conjugate FITC
  • Catalog # SA125265
  • Size 125µg
Applications
Flow Cytometry (5 µg/ml for 10^6 cells in 100 µl)
Alternate names:
Catalog # Size Price (AUD) Your Price (AUD) Qty
SA125265 208.00
Check your price
  • Goat anti-Mouse IgG1 Secondary Antibody, Alexa Fluor® 594 conjugate for IF, ICC, IHC and Flow
  • Host Goat
  • Target Species Mouse
  • Conjugate
  • Catalog # A21125
  • Size 250µL
Applications
Flow Cytometry (1-10 µg/mL), Immunofluorescence (1-10 µg/mL)
Alternate names:
Catalog # Size Price (AUD) Your Price (AUD) Qty
A21125 149.00
Check your price
  • Goat anti-Mouse IgG1 Secondary Antibody, Alexa Fluor® 680 conjugate for Western Blot, IF and ICC
  • Host Goat
  • Target Species Mouse
  • Conjugate
  • Catalog # A31562
  • Size 250µL
Applications
Immunocytochemistry (1-10 µg/mL), Immunofluorescence (1-10 µg/mL), Western Blot (0.04-0.2 µg/mL)
Alternate names:
Catalog # Size Price (AUD) Your Price (AUD) Qty
A31562 149.00
Check your price
  • Goat anti-Mouse IgG1 Secondary Antibody, PE conjugate for IF, ICC, IHC and Flow
  • Host Goat
  • Target Species Mouse
  • Conjugate PE
  • Catalog # P21129
  • Size 250µL
Applications
Flow Cytometry (1-10 µg/mL)
Alternate names:
Catalog # Size Price (AUD) Your Price (AUD) Qty
P21129 176.00
Check your price
  • Rat anti-Mouse IgG1 Secondary Antibody, Biotin conjugate for ELISA
  • Host Rat
  • Target Species Mouse
  • Conjugate Biotin
  • Catalog # 046140
  • Size 1mL
Applications
ELISA (1:1,000), Flow Cytometry (1:1,000), Immunofluorescence (1:1,000), Immunohistochemistry (1:1,000), Western Blot (1:1,000)
Alternate names:
Catalog # Size Price (AUD) Your Price (AUD) Qty
046140 179.00
Check your price
  • Rat anti-Mouse IgG1 Secondary Antibody, FITC conjugate for Western Blot, IF, IHC (P), IP and ELISA
  • Host Rat
  • Target Species Mouse
  • Conjugate
  • Catalog # 046111
  • Size 400µg
Applications
ELISA (1:20), Immunofluorescence (1:20), Immunohistochemistry (Paraffin) (1:20), Immunoprecipitation (1:20), Western Blot (1:20)
Alternate names:
Catalog # Size Price (AUD) Your Price (AUD) Qty
046111 204.00
Check your price
  • Rat anti-Mouse IgG1 Secondary Antibody, HRP conjugate for ELISA
  • Host Rat
  • Target Species Mouse
  • Conjugate HRP
  • Catalog # 046120
  • Size 1mL
Applications
ELISA (1:500-1:1,000), Immunofluorescence (1:500-1:1,000), Immunohistochemistry (Paraffin) (1:500-1:1,000), Immunoprecipitation (1:500-1:1,000), Western Blot (1:500-1:1,000)
Alternate names:
Catalog # Size Price (AUD) Your Price (AUD) Qty
046120 Not Orderable Online
Check your price
  • Goat anti-Rat IgG1 Secondary Antibody, HRP conjugate for Western Blot and ELISA
  • Host Goat
  • Target Species Rat
  • Conjugate HRP
  • Catalog # PA184708
  • Size 500µg
Applications
ELISA (1:10,000), Western Blot (1:1,000-1:10,000)
Alternate names:
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA184708 365.00
Check your price
  • Rat anti-Mouse IgG1 Secondary Antibody for Western Blot, IF, IHC (P), IP and ELISA
  • Host Rat
  • Target Species Mouse
  • Conjugate Unconjugated
  • Catalog # 046100
  • Size 500µg
Applications
ELISA (1:500-1:20,000), Immunofluorescence (1:500-1:20,000), Immunohistochemistry (Paraffin) (1:500-1:20,000), Immunoprecipitation (1:500-1:20,000), Western Blot (1:500-1:20,000)
Alternate names:
Catalog # Size Price (AUD) Your Price (AUD) Qty
046100 188.00
Check your price
  • Goat anti-Mouse IgG1 Secondary Antibody, PE-Texas Red® conjugate for Flow
  • Host Goat
  • Target Species Mouse
  • Conjugate PE-Texas Red®
  • Catalog # M32017
  • Size 500µL
Applications
Flow Cytometry (1:20-1:80)
Alternate names:
Catalog # Size Price (AUD) Your Price (AUD) Qty
M32017 256.00
Check your price
  • Goat anti-Mouse IgG1 Secondary Antibody, PE-Cyanine5 conjugate for Flow
  • Host Goat
  • Target Species Mouse
  • Conjugate PE-Cyanine5
  • Catalog # M32018
  • Size 500µL
Applications
Flow Cytometry (1:20-1:80)
Alternate names:
Catalog # Size Price (AUD) Your Price (AUD) Qty
M32018 332.00
Check your price
  • Goat anti-Mouse IgG1 Secondary Antibody, TRI-COLOR® conjugate for Flow
  • Host Goat
  • Target Species Mouse
  • Conjugate
  • Catalog # M32006
  • Size 500µL
Applications
Flow Cytometry (1:20-1:80)
Alternate names:
Catalog # Size Price (AUD) Your Price (AUD) Qty
M32006 Not Orderable Online
Check your price
  • Mouse anti-Human IgG1 Secondary Antibody for IF
  • Host Mouse
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # MH1013
  • Size 500µL
Applications
Immunofluorescence (1:500-1:20,000)
Alternate names:
Catalog # Size Price (AUD) Your Price (AUD) Qty
MH1013 152.00
Check your price
  • Goat anti-Mouse IgG1 Secondary Antibody, APC conjugate for Flow
  • Host Goat
  • Target Species Mouse
  • Conjugate
  • Catalog # A10541
  • Size 250µL
Applications
Flow Cytometry (1-10 µg/mL)
Alternate names:
Catalog # Size Price (AUD) Your Price (AUD) Qty
A10541 259.00
Check your price