1 - 15 of 15 products displayed
  • Mouse anti-Human IgG2 Fab Secondary Antibody for IHC (F) and ELISA
  • Host Mouse
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # MA134755
  • Size 100 µg
Applications
ELISA (Assay dependent), Immunohistochemistry (Frozen) (Assay dependent)
Alternate names:
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA134755 364.00
Check your price
  • Mouse anti-Human IgG2 Fd Secondary Antibody, HRP conjugate for ELISA
  • Host Mouse
  • Target Species Human
  • Conjugate HRP
  • Catalog # 050520
  • Size 500 µL
Applications
ELISA (1:500-1:1,000), Immunofluorescence (1:500-1:1,000), Immunohistochemistry (Paraffin) (1:500-1:1,000), Immunoprecipitation (1:500-1:1,000), Western Blot (1:500-1:1,000)
Alternate names:
Catalog # Size Price (AUD) Your Price (AUD) Qty
050520 149.00
Check your price
  • Sheep anti-Bovine IgG2 Secondary Antibody, HRP conjugate for ELISA
  • Host Sheep
  • Target Species Bovine
  • Conjugate HRP
  • Catalog # PA184661
  • Size 1 mg
Applications
ELISA (1/10,000-1/100,000)
Alternate names:
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA184661 359.00
Check your price
  • Mouse anti-Bovine IgG2 Secondary Antibody, HRP conjugate for ELISA
  • Host Mouse
  • Target Species Bovine
  • Conjugate HRP
  • Catalog # MA516737
  • Size 250 ug
Applications
ELISA (1:100-1:1000)
Alternate names:
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA516737 375.00
Check your price
  • Mouse anti-Human IgG2 Secondary Antibody, HRP conjugate for Western Blot, IHC and ELISA
  • Host Mouse
  • Target Species Human
  • Conjugate HRP
  • Catalog # MH1722
  • Size 500 µL
Applications
ELISA (1:1,000), Immunohistochemistry (1:1,000), Western Blot (1:1,000)
Alternate names:
Catalog # Size Price (AUD) Your Price (AUD) Qty
MH1722 208.00
Check your price
  • Mouse anti-Human IgG2 Fab Secondary Antibody for IHC (F) and ELISA
  • Host Mouse
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # MA516715
  • Size 200 ug
Applications
ELISA (Assay Dependent), Immunohistochemistry (Frozen) (Assay Dependent)
Alternate names:
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA516715 425.00
Check your price
  • Mouse anti-Human IgG2 Secondary Antibody, Biotin conjugate for ELISA
  • Host Mouse
  • Target Species Human
  • Conjugate Biotin
  • Catalog # 053540
  • Size 500 µL
Applications
ELISA (1:500-1:1,000), Immunofluorescence (1:500-1:1,000), Immunohistochemistry (Paraffin) (1:500-1:1,000), Immunoprecipitation (1:500-1:1,000), Western Blot (1:500-1:1,000)
Alternate names:
Catalog # Size Price (AUD) Your Price (AUD) Qty
053540 129.00
Check your price
  • Mouse anti-Human IgG2 Secondary Antibody, AP conjugate for Western Blot, IHC (P), IP and ELISA
  • Host Mouse
  • Target Species Human
  • Conjugate AP
  • Catalog # 053522
  • Size 500 µL
Applications
ELISA (1:500), Immunofluorescence (1:500), Immunohistochemistry (Paraffin) (1:500), Immunoprecipitation (1:500), Western Blot (1:500)
Alternate names:
Catalog # Size Price (AUD) Your Price (AUD) Qty
053522 139.00
Check your price
  • Mouse anti-Human IgG2 Secondary Antibody for Western Blot, IF, IHC (P), IP and ELISA
  • Host Mouse
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # 053500
  • Size 500 µg
Applications
ELISA (1:500-1:20,000), Immunofluorescence (1:500-1:20,000), Immunohistochemistry (Paraffin) (1:500-1:20,000), Immunoprecipitation (1:500-1:20,000), Western Blot (1:500-1:20,000)
Alternate names:
Catalog # Size Price (AUD) Your Price (AUD) Qty
053500 189.00
Check your price
  • Mouse anti-Human IgG4 Fc Secondary Antibody, HRP conjugate for IHC (F), IHC (P) and ELISA
  • Host Mouse
  • Target Species Human
  • Conjugate HRP
  • Catalog # MA182201
  • Size 200 µg
Applications
ELISA (1:25,600-1:51,200), Immunohistochemistry (Frozen) (1/40-1/80), Immunohistochemistry (Paraffin) (Assay Dependent)
Alternate names:
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA182201 840.00
Check your price
  • Mouse anti-Human IgG4 Secondary Antibody, HRP conjugate for IHC (P) and ELISA
  • Host Mouse
  • Target Species Human
  • Conjugate HRP
  • Catalog # MA134437
  • Size 100 µg
Applications
ELISA (1:25,600-1:51,200), Immunohistochemistry (Paraffin) (Assay Dependent)
Alternate names:
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA134437 499.00
Check your price
  • Mouse anti-Human IgG2 Secondary Antibody for ELISA
  • Host Mouse
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # MA183241
  • Size 500 µg
Applications
ELISA (Assay Dependent)
Alternate names:
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA183241 212.00
Check your price
  • Goat anti-Rabbit IgG F(ab')2 Secondary Antibody, FITC conjugate for IF, ICC, IHC, Flow and IP
  • Host Goat
  • Target Species Rabbit
  • Conjugate FITC
  • Catalog # 31573
  • Size 2 mg
Applications
Flow Cytometry (1:50-1:200), Immunocytochemistry (1:50-1:200), Immunofluorescence (1:50-1:200), Immunohistochemistry (1:50-1:200), Immunoprecipitation (1:50-1:200)
Alternate names:
Catalog # Size Price (AUD) Your Price (AUD) Qty
31573 141.00
Check your price
  • Mouse anti-Human IgA Secondary Antibody for Western Blot and ELISA
  • Host Mouse
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # SA172510
  • Size 100 µg
Applications
ELISA (1 µg/ml), Western Blot (1 µg/ml)
Alternate names:
Catalog # Size Price (AUD) Your Price (AUD) Qty
SA172510 188.00
Check your price
  • Mouse anti-Human IgM Secondary Antibody for ELISA
  • Host Mouse
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # SA172512
  • Size 100 µg
Applications
ELISA (1-5 µg/ml)
Alternate names:
Catalog # Size Price (AUD) Your Price (AUD) Qty
SA172512 199.00
Check your price