1 - 5 of 5 products displayed
  • CADM4 Polyclonal Antibody for IHC (P)
  • Host Rabbit
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # PA533378
  • Size 50ug
Applications
Immunohistochemistry (Paraffin) (10-15 µg/ml)
Alternate names:
TSLC1-like protein 2; Nectin-like protein 4; Immunoglobulin superfamily member 4C
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA533378 349.00
Check your price
  • CADM4 Monoclonal Antibody for Western Blot, IF, ICC and Flow
  • Host Rat
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # MA524141
  • Size 100µg
Applications
Flow Cytometry (2.5 µg/10^6 cells), Immunocytochemistry (8-25 µg/ml), Immunofluorescence (8-25 µg/ml), Western Blot (1 µg/ml)
Alternate names:
TSLC1-like protein 2; Nectin-like protein 4; Immunoglobulin superfamily member 4C
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA524141 275.00
Check your price
  • CADM4 Polyclonal Antibody for Western Blot
  • Host Goat
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # PA518838
  • Size 100ug
Applications
Western Blot (0.03-0.1 ug/ml)
Alternate names:
TSLC1-like protein 2; Nectin-like protein 4; Immunoglobulin superfamily member 4C
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA518838 349.00
Check your price
  • CADM4 Polyclonal Antibody for Western Blot and IHC
  • Host Goat
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # PA518804
  • Size 100ug
Applications
Immunohistochemistry (2-5µg/ml), Western Blot (0.1-0.3 ug/ml)
Alternate names:
TSLC1-like protein 2; Nectin-like protein 4; Immunoglobulin superfamily member 4C
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA518804 349.00
Check your price
  • CADM4 Polyclonal Antibody for Western Blot and Flow
  • Host Goat
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # PA548046
  • Size 100µg
Applications
Flow Cytometry (2.5 µg/10^6 cells), Western Blot (0.1 µg/ml)
Alternate names:
TSLC1-like protein 2; Nectin-like protein 4; Immunoglobulin superfamily member 4C
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA548046 309.00
Check your price