1 - 7 of 7 products displayed
  • CK1 delta Polyclonal Antibody for Western Blot, IF, ICC and IHC (P)
  • Host Rabbit
  • Target Species Human, Mouse
  • Conjugate Unconjugated
  • Catalog # PA532129
  • Size 100µL
Applications
Immunocytochemistry (1:100-1:1000), Immunofluorescence (1:100-1:1000), Immunohistochemistry (Paraffin) (1:100-1:1000), Western Blot (1:500-1:3000)
Alternate names:
Tau-protein kinase CSNK1D
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA532129 349.00
Check your price
  • CK1 delta Polyclonal Antibody for IHC (P) and ELISA
  • Host Goat
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # PA128034
  • Size 50µL
Applications
ELISA (Assay-dependent), Immunohistochemistry (Paraffin) (1:200)
Alternate names:
Tau-protein kinase CSNK1D
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA128034 349.00
Check your price
  • CK1 gamma-1 Polyclonal Antibody for Western Blot, IF and IHC (P)
  • Host Rabbit
  • Target Species Human, Mouse, Rat
  • Conjugate Unconjugated
  • Catalog # PA536284
  • Size 100µl
Applications
Immunofluorescence (1:50-1:200), Immunohistochemistry (Paraffin) (1:50-1:200), Western Blot (1:500-1:1000)
Alternate names:
Casein kinase I isoform gamma-1, CKI-gamma 1, CSNK1G1, KC1G1, CK1G1, CKI gamma1, CK-I gamma1, CKI- gamma1, CKI gamma-1, CKI gamma1, CK I gamma1, CKI gamma1, CKI gamma 1, CKI gamma 1, CKI-gamma-1
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA536284 309.00
Check your price
  • Phospho-CK1 alpha (Thr321) Polyclonal Antibody for Western Blot and IHC (P)
  • Host Rabbit
  • Target Species Human, Mouse, Rat
  • Conjugate Unconjugated
  • Catalog # PA536790
  • Size 100µl
Applications
Immunohistochemistry (Paraffin) (1:50-1:200), Western Blot (1:500-1:1000)
Alternate names:
CK1
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA536790 349.00
Check your price
  • CK1 delta Polyclonal Antibody for Western Blot and IHC (P)
  • Host Rabbit
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # PA514297
  • Size 400µL
Applications
Immunohistochemistry (1:50-1:100), Western Blot (1:1000)
Alternate names:
Tau-protein kinase CSNK1D
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA514297 349.00
Check your price
  • CK1 gamma Polyclonal Antibody for IHC (P)
  • Host Rabbit
  • Target Species Human, Mouse, Rat
  • Conjugate Unconjugated
  • Catalog # PA539735
  • Size 100µg
Applications
Immunohistochemistry (Paraffin) (1:50-1:100)
Alternate names:
Casein kinase I isoform gamma-1, CKI-gamma 1, CSNK1G1, KC1G1, CK1G1
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA539735 309.00
Check your price
  • Phospho-CK1 alpha (Tyr294) Polyclonal Antibody for Western Blot and IHC (P)
  • Host Rabbit
  • Target Species Human, Mouse, Rat
  • Conjugate Unconjugated
  • Catalog # PA539733
  • Size 100µg
Applications
Immunohistochemistry (Paraffin) (1:50-1:100), Western Blot (1:500-1:1000)
Alternate names:
CK1
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA539733 309.00
Check your price