1 - 30 of 30 products displayed
  • IKK epsilon Polyclonal Antibody for Western Blot and IHC
  • Host Rabbit
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # PA519964
  • Size 100 ug
Applications
Immunohistochemistry (10 µg/ml), Western Blot (1 µg/ml)
Alternate names:
Inducible I kappa-B kinase
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA519964 349.00
Check your price
  • IKK epsilon Polyclonal Antibody for Western Blot and IHC
  • Host Rabbit
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # PA515439
  • Size 400 µL
Applications
Immunohistochemistry (1:50-1:100), Western Blot (1:1000)
Alternate names:
Inducible I kappa-B kinase
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA515439 349.00
Check your price
  • IKK epsilon Polyclonal Antibody for Western Blot
  • Host Rabbit
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # PA125443
  • Size 50 µg
Applications
Western Blot (1-2 µg/ml)
Alternate names:
Inducible I kappa-B kinase
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA125443 375.00
Check your price
  • IKK epsilon Monoclonal Antibody for Western Blot and IP
  • Host Rabbit
  • Target Species Human, Non-human primate
  • Conjugate Unconjugated
  • Catalog # MA514817
  • Size 100 µL
Applications
Immunoprecipitation (1:100), Western Blot (1:1000)
Alternate names:
Inducible I kappa-B kinase
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA514817 275.00
Check your price
  • IKK epsilon Polyclonal Antibody for Western Blot
  • Host Rabbit
  • Target Species Human, Mouse, Rat
  • Conjugate Unconjugated
  • Catalog # PA517221
  • Size 100 µL
Applications
Western Blot (1:1000)
Alternate names:
Inducible I kappa-B kinase
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA517221 275.00
Check your price
  • IKK epsilon Monoclonal Antibody for Western Blot
  • Host Mouse
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # MA515377
  • Size 100 µL
Applications
Western Blot (1/500-1/2000)
Alternate names:
Inducible I kappa-B kinase
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA515377 349.00
Check your price
  • IKK epsilon Polyclonal Antibody for Western Blot, IF, ICC and IHC (P)
  • Host Rabbit
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # PA529294
  • Size 100 µL
Applications
Immunocytochemistry (1:100-1:1000), Immunofluorescence (1:100-1:1000), Immunohistochemistry (Paraffin) (1:100-1:1000), Western Blot (1:500-1:3000)
Alternate names:
Inducible I kappa-B kinase
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA529294 349.00
Check your price
  • IKK epsilon Polyclonal Antibody for Western Blot, IF and ICC
  • Host Rabbit
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # PA528264
  • Size 100 µL
Applications
Western Blot (1:5000-1:20,000)
Alternate names:
Inducible I kappa-B kinase
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA528264 349.00
Check your price
  • IKK alpha Oligoclonal Antibody for Western Blot, IF and ICC
  • Host Rabbit
  • Target Species Human, Non-human primate
  • Conjugate Unconjugated
  • Catalog # 710719
  • Size 100 µg
Applications
Immunocytochemistry (1 µg/ml), Immunofluorescence (1 µg/ml), Western Blot (1-2 µg/ml)
Alternate names:
Transcription factor 16; Nuclear factor NF-kappa-B inhibitor kinase alpha; I-kappa-B kinase 1; Conserved helix-loop-helix ubiquitous kinase
Catalog # Size Price (AUD) Your Price (AUD) Qty
710719 325.00
Check your price
  • IKK alpha Polyclonal Antibody for Western Blot and ICC
  • Host Rabbit
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # PA519915
  • Size 100 ug
Applications
Immunocytochemistry (10 µg/ml), Western Blot (0.5 µg/ml)
Alternate names:
Transcription factor 16; Nuclear factor NF-kappa-B inhibitor kinase alpha; I-kappa-B kinase 1; Conserved helix-loop-helix ubiquitous kinase
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA519915 349.00
Check your price
  • IKK alpha Polyclonal Antibody for Western Blot, IF and ICC
  • Host Rabbit
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # PA519914
  • Size 100 µg
Applications
Immunocytochemistry (10 µg/ml), Immunofluorescence (20 ug/ml), Western Blot (0.5 µg/ml)
Alternate names:
Transcription factor 16; Nuclear factor NF-kappa-B inhibitor kinase alpha; I-kappa-B kinase 1; Conserved helix-loop-helix ubiquitous kinase
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA519914 349.00
Check your price
  • IKK alpha Polyclonal Antibody for Western Blot, IF and ICC
  • Host Rabbit
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # PA519897
  • Size 100 ug
Applications
Immunocytochemistry (1 µg/ml), Immunofluorescence (20 µg/ml), Western Blot (1 µg/ml)
Alternate names:
Transcription factor 16; Nuclear factor NF-kappa-B inhibitor kinase alpha; I-kappa-B kinase 1; Conserved helix-loop-helix ubiquitous kinase
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA519897 349.00
Check your price
  • IKK gamma Polyclonal Antibody for Western Blot and ICC
  • Host Rabbit
  • Target Species Human, Mouse, Rat
  • Conjugate Unconjugated
  • Catalog # PA519965
  • Size 100 ug
Applications
Immunocytochemistry (5 µg/ml), Western Blot (1 µg/ml)
Alternate names:
NF-kappa-B essential modifier; Inhibitor of nuclear factor kappa-B kinase subunit gamma; IkB kinase-associated protein 1; FIP-3
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA519965 349.00
Check your price
  • IKK beta Polyclonal Antibody for Western Blot, IF and ICC
  • Host Rabbit
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # PA519916
  • Size 100 ug
Applications
Immunocytochemistry (10 µg/ml), Immunofluorescence (10 ug/ml), Western Blot (0.5 µg/ml)
Alternate names:
Nuclear factor NF-kappa-B inhibitor kinase beta; I-kappa-B kinase 2
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA519916 349.00
Check your price
  • IKK alpha Monoclonal Antibody for Western Blot, IF, ICC, IHC (F), IHC (P), Flow and IP
  • Host Mouse
  • Target Species Human, Mouse, Non-human primate
  • Conjugate Unconjugated
  • Catalog # MA516157
  • Size 100 ug
Applications
Flow Cytometry (0.25-0.5 µg/10^6 cells), Immunocytochemistry (1:10), Immunofluorescence (1:10), Immunohistochemistry (Paraffin) (5 µg/ml), Immunoprecipitation (1-2 µg/ml), Western Blot (1 µg/ml)
Alternate names:
Transcription factor 16; Nuclear factor NF-kappa-B inhibitor kinase alpha; I-kappa-B kinase 1; Conserved helix-loop-helix ubiquitous kinase
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA516157 349.00
Check your price
  • IKK gamma Monoclonal Antibody for Western Blot
  • Host Mouse
  • Target Species Human, Rat
  • Conjugate Unconjugated
  • Catalog # MA515155
  • Size 100 µL
Applications
Western Blot (1:1000)
Alternate names:
NF-kappa-B essential modifier; Inhibitor of nuclear factor kappa-B kinase subunit gamma; IkB kinase-associated protein 1; FIP-3
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA515155 275.00
Check your price
  • IKK gamma Polyclonal Antibody for Western Blot, IF, ICC and IHC (P)
  • Host Rabbit
  • Target Species Human, Mouse, Rat
  • Conjugate Unconjugated
  • Catalog # PA528606
  • Size 100 µL
Applications
Immunocytochemistry (1:100-1:1000), Immunofluorescence (1:100-1:1000), Immunohistochemistry (Paraffin) (1:100-1:1000), Western Blot (1:500-1:3000)
Alternate names:
NF-kappa-B essential modifier; Inhibitor of nuclear factor kappa-B kinase subunit gamma; IkB kinase-associated protein 1; FIP-3
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA528606 349.00
Check your price
  • IKK gamma Polyclonal Antibody for Western Blot, IF, ICC and IHC (P)
  • Host Rabbit
  • Target Species Human, Mouse
  • Conjugate Unconjugated
  • Catalog # PA527335
  • Size 100 µL
Applications
Immunocytochemistry (1:100-1:1000), Immunofluorescence (1:100-1:1000), Immunohistochemistry (Paraffin) (1:100-1:1000), Western Blot (1:500-1:3000)
Alternate names:
NF-kappa-B essential modifier; Inhibitor of nuclear factor kappa-B kinase subunit gamma; IkB kinase-associated protein 1; FIP-3
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA527335 349.00
Check your price
  • IKK beta Monoclonal Antibody for Western Blot
  • Host Mouse
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # MA511177
  • Size 400 µL
Applications
Western Blot (1:100-1:500)
Alternate names:
Nuclear factor NF-kappa-B inhibitor kinase beta; I-kappa-B kinase 2
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA511177 349.00
Check your price
  • Phospho-IKK alpha (Thr23) Polyclonal Antibody for Western Blot and IHC (P)
  • Host Rabbit
  • Target Species Human, Mouse, Rat
  • Conjugate Unconjugated
  • Catalog # PA536652
  • Size 100 µl
Applications
Immunohistochemistry (Paraffin) (1:50-1:200), Western Blot (1:500-1:1000)
Alternate names:
Transcription factor 16; Nuclear factor NF-kappa-B inhibitor kinase alpha; I-kappa-B kinase 1; Conserved helix-loop-helix ubiquitous kinase
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA536652 349.00
Check your price
  • Phospho-IKK alpha (Ser176, Ser180) Polyclonal Antibody for Western Blot, IF, ICC, Flow and ChIP
  • Host Rabbit
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # 44714
  • Size 10 blots
Applications
ChIP assay (10µl), Flow Cytometry (3-5 µg/million cells), Immunocytochemistry (1 µg/ml), Immunofluorescence (1 µg/ml), Western Blot (0.1-1.0 ug/ml)
Alternate names:
Transcription factor 16; Nuclear factor NF-kappa-B inhibitor kinase alpha; I-kappa-B kinase 1; Conserved helix-loop-helix ubiquitous kinase
Catalog # Size Price (AUD) Your Price (AUD) Qty
44714 305.00
Check your price
  • Phospho-IKK beta (Tyr188) Polyclonal Antibody for Western Blot and IHC (P)
  • Host Rabbit
  • Target Species Human, Mouse, Rat
  • Conjugate Unconjugated
  • Catalog # PA536653
  • Size 100 µl
Applications
Immunohistochemistry (Paraffin) (1:50-1:200), Western Blot (1:500-1:1000)
Alternate names:
Nuclear factor NF-kappa-B inhibitor kinase beta; I-kappa-B kinase 2
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA536653 349.00
Check your price
  • IKK beta Monoclonal Antibody for Western Blot
  • Host Rabbit
  • Target Species Human, Mouse, Non-human primate, Rat
  • Conjugate Unconjugated
  • Catalog # MA515080
  • Size 100 µL
Applications
Western Blot (1:1000)
Alternate names:
Nuclear factor NF-kappa-B inhibitor kinase beta; I-kappa-B kinase 2
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA515080 275.00
Check your price
  • IKK beta Polyclonal Antibody for Western Blot, IF, ICC and IHC (P)
  • Host Rabbit
  • Target Species Human, Rat
  • Conjugate Unconjugated
  • Catalog # PA521683
  • Size 100 µL
Applications
Immunocytochemistry (1:100-1:1000), Immunofluorescence (1:100-1:1000), Immunohistochemistry (Paraffin) (1:100-1:1000), Western Blot (1:500-1:3000)
Alternate names:
Nuclear factor NF-kappa-B inhibitor kinase beta; I-kappa-B kinase 2
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA521683 349.00
Check your price
  • Phospho-IKK alpha/beta (Ser176, Ser177) Monoclonal Antibody for Western Blot
  • Host Rabbit
  • Target Species Bovine, Human, Mouse, Non-human primate, Rat
  • Conjugate Unconjugated
  • Catalog # MA514857
  • Size 100 µL
Applications
Western Blot (1:1000)
Alternate names:
Nuclear factor NF-kappa-B inhibitor kinase beta; I-kappa-B kinase 2; Transcription factor 16; Nuclear factor NF-kappa-B inhibitor kinase alpha; I-kappa-B kinase 1; Conserved helix-loop-helix ubiquitous kinase
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA514857 309.00
Check your price
  • Phospho-IKK alpha/beta (Ser180, Ser181) Polyclonal Antibody for Western Blot and IHC (P)
  • Host Rabbit
  • Target Species Human, Mouse, Rat
  • Conjugate Unconjugated
  • Catalog # PA536718
  • Size 100 µl
Applications
Immunohistochemistry (Paraffin) (1:50-1:200), Western Blot (1:500-1:1000)
Alternate names:
Nuclear factor NF-kappa-B inhibitor kinase beta; I-kappa-B kinase 2; Transcription factor 16; Nuclear factor NF-kappa-B inhibitor kinase alpha; I-kappa-B kinase 1; Conserved helix-loop-helix ubiquitous kinase
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA536718 349.00
Check your price
  • Phospho-IKK alpha/beta (Ser176, Ser180) Oligoclonal Antibody for IF and ICC
  • Host Rabbit
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # 710676
  • Size 100 µg
Applications
Immunocytochemistry (1-2 µg/ml), Immunofluorescence (1-2 µg/ml)
Alternate names:
Transcription factor 16; Nuclear factor NF-kappa-B inhibitor kinase alpha; I-kappa-B kinase 1; Conserved helix-loop-helix ubiquitous kinase; Nuclear factor NF-kappa-B inhibitor kinase beta; I-kappa-B kinase 2
Catalog # Size Price (AUD) Your Price (AUD) Qty
710676 325.00
Check your price
  • Phospho-IKK alpha/beta (Ser176, Ser180) Monoclonal Antibody for IF and ICC
  • Host Rabbit
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # 701643
  • Size 100 µg
Applications
Immunocytochemistry (1-2 µg/ml), Immunofluorescence (1-2 µg/ml)
Alternate names:
Transcription factor 16; Nuclear factor NF-kappa-B inhibitor kinase alpha; I-kappa-B kinase 1; Conserved helix-loop-helix ubiquitous kinase; Nuclear factor NF-kappa-B inhibitor kinase beta; I-kappa-B kinase 2
Catalog # Size Price (AUD) Your Price (AUD) Qty
701643 325.00
Check your price
  • Phospho-IKK alpha (Thr23) Polyclonal Antibody for Western Blot and IHC (P)
  • Host Rabbit
  • Target Species Human, Mouse, Rat
  • Conjugate Unconjugated
  • Catalog # PA537524
  • Size 100 µl
Applications
Immunohistochemistry (Paraffin) (1:50-1:100), Western Blot (1:500-1:1000)
Alternate names:
Transcription factor 16; Nuclear factor NF-kappa-B inhibitor kinase alpha; I-kappa-B kinase 1; Conserved helix-loop-helix ubiquitous kinase
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA537524 309.00
Check your price
  • Phospho-IKK gamma (Ser376) Polyclonal Antibody for IHC (P)
  • Host Rabbit
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # PA537766
  • Size 100 µl
Applications
Immunohistochemistry (Paraffin) (1:50-1:100)
Alternate names:
NF-kappa-B essential modifier; Inhibitor of nuclear factor kappa-B kinase subunit gamma; IkB kinase-associated protein 1; FIP-3
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA537766 309.00
Check your price