1 - 25 of 25 products displayed
  • IL-22 Polyclonal Antibody for Western Blot, ELISA and Neu
  • Host Goat
  • Target Species Mouse
  • Conjugate Unconjugated
  • Catalog # PA547782
  • Size 100µg
Applications
ELISA (0.2-0.8 µg/ml (Detection), 1-4 µg/ml (Capture)), Neutralization (0.5-2.5 µg/ml), Western Blot (0.1 µg/ml)
Alternate names:
Interleukin-22a; IL-TIF alpha; IL-10-related T-cell-derived-inducible factor
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA547782 349.00
Check your price
  • IL-22 Polyclonal Antibody for IHC (P) and ELISA
  • Host Rabbit
  • Target Species Human, Mouse
  • Conjugate Unconjugated
  • Catalog # PA121357
  • Size 50µg
Applications
ELISA (1:100,000), Immunocytochemistry (1:200), Immunofluorescence (1:200), Immunohistochemistry (Paraffin) (1:500)
Alternate names:
Interleukin-22a; IL-TIF alpha; IL-10-related T-cell-derived-inducible factor; Cytokine Zcyto18
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA121357 399.00
Check your price
  • IL-22 Polyclonal Antibody for IF, ICC, IHC (P) and ELISA
  • Host Goat
  • Target Species Dog, Human, Mouse, Rat
  • Conjugate Unconjugated
  • Catalog # PA121356
  • Size 50µg
Applications
ELISA (1:100,000), Immunocytochemistry (1:300), Immunofluorescence (1:300), Immunohistochemistry (Paraffin) (Assay-dependent)
Alternate names:
Interleukin-22a; IL-TIF alpha; IL-10-related T-cell-derived-inducible factor; Cytokine Zcyto18
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA121356 399.00
Check your price
  • IL-22 Polyclonal Antibody for IHC (P)
  • Host Rabbit
  • Target Species Human, Mouse
  • Conjugate Unconjugated
  • Catalog # PA121358
  • Size 50µg
Applications
Immunohistochemistry (Paraffin) (1:500)
Alternate names:
Interleukin-22a; IL-TIF alpha; IL-10-related T-cell-derived-inducible factor; Cytokine Zcyto18
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA121358 475.00
Check your price
  • IL-22 Polyclonal Antibody for Western Blot
  • Host Rabbit
  • Target Species Human, Mouse
  • Conjugate Unconjugated
  • Catalog # PA534944
  • Size 100µL
Applications
Western Blot (1:5000-1:20,000)
Alternate names:
Interleukin-22a; IL-TIF alpha; IL-10-related T-cell-derived-inducible factor; Cytokine Zcyto18
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA534944 349.00
Check your price
  • IL-22 Polyclonal Antibody for Western Blot and IHC (P)
  • Host Rabbit
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # PA534753
  • Size 100µL
Applications
Immunohistochemistry (Paraffin) (1:100-1:1000), Western Blot (1:500-1:3000)
Alternate names:
IL-10-related T-cell-derived-inducible factor; Cytokine Zcyto18
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA534753 349.00
Check your price
  • IL-22 Oligoclonal Antibody for Western Blot
  • Host Rabbit
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # 710084
  • Size 100µg
Applications
Western Blot (1-5 ug/ml)
Alternate names:
IL-10-related T-cell-derived-inducible factor; Cytokine Zcyto18
Catalog # Size Price (AUD) Your Price (AUD) Qty
710084 309.00
Check your price
  • IL22RA1 Polyclonal Antibody for Western Blot
  • Host Rabbit
  • Target Species Human, Mouse
  • Conjugate Unconjugated
  • Catalog # PA130723
  • Size 100µg
Applications
Western Blot (0.5-1 µg/ml)
Alternate names:
ZcytoR11; Cytokine receptor family 2 member 9; Cytokine receptor class-II member 9
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA130723 449.00
Check your price
  • IL-20 Polyclonal Antibody for Western Blot and Neu
  • Host Goat
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # PA547067
  • Size 100µg
Applications
Neutralization (0.1-0.4 µg/ml), Western Blot (0.1 µg/ml)
Alternate names:
Cytokine Zcyto10
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA547067 349.00
Check your price
  • IL-6 Polyclonal Antibody for Western Blot
  • Host Rabbit
  • Target Species Rat
  • Conjugate Unconjugated
  • Catalog # PR627
  • Size 1mg
Applications
Western Blot (Assay Dependent)
Alternate names:
IL-6; Interleukin 6 (interferon, beta 2); interleukin-6
Catalog # Size Price (AUD) Your Price (AUD) Qty
PR627 349.00
Check your price
  • IL-6 Polyclonal Antibody for IHC (P), ELISA, IB and Neu
  • Host Rabbit
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # P620
  • Size 1mg
Applications
ELISA (Assay Dependent), Immunoblot (Assay Dependent), Immunohistochemistry (Paraffin) (Assay Dependent), Neutralization (Assay Dependent)
Alternate names:
Interferon beta-2; Hybridoma growth factor; CTL differentiation factor; B-cell stimulatory factor 2
Catalog # Size Price (AUD) Your Price (AUD) Qty
P620 275.00
Check your price
  • IL-6 Polyclonal Antibody for Western Blot and ELISA
  • Host Rabbit
  • Target Species Mouse
  • Conjugate Unconjugated
  • Catalog # PM626
  • Size 500ug
Applications
ELISA (Assay Dependent), Western Blot (Assay Dependent)
Alternate names:
Interleukin HP-1; B-cell hybridoma growth factor
Catalog # Size Price (AUD) Your Price (AUD) Qty
PM626 325.00
Check your price
  • IL22RA1 Polyclonal Antibody for Western Blot, IF and ICC
  • Host Rabbit
  • Target Species Human, Rat
  • Conjugate Unconjugated
  • Catalog # PA519987
  • Size 100ug
Applications
Immunocytochemistry (10 µg/ml), Immunofluorescence (10 ug/ml), Western Blot (1 µg/ml)
Alternate names:
ZcytoR11; Cytokine receptor family 2 member 9; Cytokine receptor class-II member 9
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA519987 349.00
Check your price
  • IL22RA1 Polyclonal Antibody for Western Blot
  • Host Rabbit
  • Target Species Human, Rat
  • Conjugate Unconjugated
  • Catalog # PA519986
  • Size 100ug
Applications
Western Blot (1 µg/ml)
Alternate names:
ZcytoR11; Cytokine receptor family 2 member 9; Cytokine receptor class-II member 9
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA519986 349.00
Check your price
  • IL22RA2 Polyclonal Antibody for IF, ICC, IHC (P), Flow and ELISA
  • Host Goat
  • Target Species Human, Mouse, Rat
  • Conjugate Unconjugated
  • Catalog # PA121359
  • Size 100µg
Applications
ELISA (1:100,000), Flow Cytometry (Assay dependent), Immunocytochemistry (1:300), Immunofluorescence (1:300), Immunohistochemistry (Paraffin) (Assay-dependent)
Alternate names:
ZcytoR16; Interleukin-22-binding protein; Cytokine receptor family type 2, soluble 1; Cytokine receptor family 2 member 10; Cytokine receptor class-II member 10
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA121359 425.00
Check your price
  • IL22RA2 Polyclonal Antibody for Western Blot and IHC (P)
  • Host Rabbit
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # PA521490
  • Size 100µL
Applications
Immunohistochemistry (Paraffin) (1:100-1:1000), Western Blot (1:100-1:1000)
Alternate names:
ZcytoR16; Interleukin-22-binding protein; Cytokine receptor family type 2, soluble 1; Cytokine receptor family 2 member 10; Cytokine receptor class-II member 10
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA521490 349.00
Check your price
  • IL-20 Polyclonal Antibody for Western Blot and Neu
  • Host Goat
  • Target Species Mouse
  • Conjugate Unconjugated
  • Catalog # PA547092
  • Size 100µg
Applications
Neutralization (0.2-0.6 µg/ml), Western Blot (0.1 µg/ml)
Alternate names:
Cytokine Zcyto10
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA547092 349.00
Check your price
  • IL22RA1 Polyclonal Antibody for IHC (P) and ELISA
  • Host Goat
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # PA121361
  • Size 100µg
Applications
ELISA (1:100,000), Immunohistochemistry (Paraffin) (1:300)
Alternate names:
ZcytoR11; Cytokine receptor family 2 member 9; Cytokine receptor class-II member 9
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA121361 399.00
Check your price
  • TSC22D3 Polyclonal Antibody for Western Blot, IF, ICC and IHC
  • Host Rabbit
  • Target Species Human, Mouse
  • Conjugate Unconjugated
  • Catalog # PA534506
  • Size 100ug
Applications
Immunocytochemistry (20 ug/ml), Immunofluorescence (20 ug/ml), Immunohistochemistry (5 ug/ml), Western Blot (1-2 ug/mL)
Alternate names:
TSC-22-related protein; TSC-22-like protein; Glucocorticoid-induced leucine zipper protein; Delta sleep-inducing peptide immunoreactor; DSIP-immunoreactive peptide; TSC22-related-inducible leucine zipper 3
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA534506 349.00
Check your price
  • Phospho-DOK2 (Tyr142) Polyclonal Antibody for Western Blot
  • Host Rabbit
  • Target Species Mouse
  • Conjugate Unconjugated
  • Catalog # 44264A
  • Size 10blots
Applications
Western Blot (Assay Dependent)
Alternate names:
p56(dok-2); IL-four receptor-interacting protein; Downstream of tyrosine kinase 2; Dok-related protein
Catalog # Size Price (AUD) Your Price (AUD) Qty
44264A 309.00
Check your price
  • MDC Polyclonal Antibody for Western Blot
  • Host Rabbit
  • Target Species Mouse, Rat
  • Conjugate Unconjugated
  • Catalog # PA122530
  • Size 50µg
Applications
Western Blot (1:2000)
Alternate names:
Small-inducible cytokine A22; CC chemokine ABCD-1; Activated B and dendritic cell-derived
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA122530 349.00
Check your price
  • PRMT2 Polyclonal Antibody for Western Blot and IHC
  • Host Rabbit
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # PA511114
  • Size 400µL
Applications
Immunohistochemistry (1:50-1:100), Western Blot (1:1000)
Alternate names:
Histone-arginine N-methyltransferase PRMT2
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA511114 349.00
Check your price
  • RORC Polyclonal Antibody for IHC (P)
  • Host Rabbit
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # PA534166
  • Size 50ug
Applications
Immunohistochemistry (Paraffin) (22 µg/ml)
Alternate names:
Retinoid-related orphan receptor-gamma; RAR-related orphan receptor C; Nuclear receptor subfamily 1 group F member 3; Nuclear receptor RZR-gamma
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA534166 349.00
Check your price
  • IL-22 Receptor Synthetic Peptide for BLOCK and Ctrl
  • Host N/A
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # PEP0110
  • Size 50ug
Applications
Blocking Assay (Assay Dependent), Control (Assay Dependent)
Alternate names:
Catalog # Size Price (AUD) Your Price (AUD) Qty
PEP0110 85.75
Check your price
  • IL-22 Receptor Synthetic Peptide for BLOCK and Ctrl
  • Host N/A
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # PEP0111
  • Size 50ug
Applications
Blocking Assay (Assay Dependent), Control (Assay Dependent)
Alternate names:
Catalog # Size Price (AUD) Your Price (AUD) Qty
PEP0111 85.75
Check your price