1 - 30 of 30 products displayed
  • IL-23 Oligoclonal Antibody for Western Blot
  • Host Rabbit
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # 710259
  • Size 100 µg
Applications
Western Blot (1-3 ug/ml)
Alternate names:
Interleukin-23 subunit p19
Catalog # Size Price (AUD) Your Price (AUD) Qty
710259 309.00
Check your price
  • IL-23 Monoclonal Antibody for Western Blot
  • Host Rabbit
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # 701235
  • Size 100 µg
Applications
Western Blot (1-2 ug/ml)
Alternate names:
Interleukin-23 subunit p19
Catalog # Size Price (AUD) Your Price (AUD) Qty
701235 309.00
Check your price
  • IL-23 Polyclonal Antibody for Western Blot, IF, ICC and IHC
  • Host Rabbit
  • Target Species Human, Mouse
  • Conjugate Unconjugated
  • Catalog # PA520239
  • Size 100 ug
Applications
Immunocytochemistry (20 ug/ml), Immunofluorescence (20 ug/ml), Immunohistochemistry (2 µg/ml), Western Blot (1-2 ug/ml)
Alternate names:
Interleukin-23 subunit p19
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA520239 349.00
Check your price
  • IL-23 Polyclonal Antibody for Western Blot, IF and ICC
  • Host Rabbit
  • Target Species Human, Mouse
  • Conjugate Unconjugated
  • Catalog # PA520238
  • Size 100 ug
Applications
Immunocytochemistry (10 µg/ml), Immunofluorescence (20 ug/ml), Western Blot (1 µg/ml)
Alternate names:
Interleukin-23 subunit p19
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA520238 349.00
Check your price
  • IL-23 Monoclonal Antibody, Alexa Fluor® 488 conjugate for Flow
  • Host Mouse
  • Target Species Human
  • Conjugate Alexa Fluor® 488
  • Catalog # MA523613
  • Size 100 Tests
Applications
Flow Cytometry (5 µl/10^6 cells)
Alternate names:
Interleukin-23 subunit p19
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA523613 289.00
Check your price
  • IL-23 Monoclonal Antibody, PE conjugate for Flow
  • Host Mouse
  • Target Species Human
  • Conjugate PE
  • Catalog # MA523644
  • Size 100 Tests
Applications
Flow Cytometry (10 µl/10^6 cells)
Alternate names:
Interleukin-23 subunit p19
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA523644 309.00
Check your price
  • IL-23 Polyclonal Antibody for Western Blot and Neu
  • Host Goat
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # PA546913
  • Size 50 µg
Applications
Neutralization (0.2-0.8 µg/ml), Western Blot (1 µg/ml)
Alternate names:
Interleukin-23 subunit p19
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA546913 375.00
Check your price
  • IL-23 Monoclonal Antibody, PE conjugate for Flow
  • Host Rat
  • Target Species Mouse
  • Conjugate PE
  • Catalog # MA523617
  • Size 100 Tests
Applications
Flow Cytometry (10 µl/10^6 cells)
Alternate names:
Interleukin-23 subunit p19
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA523617 299.00
Check your price
  • IL-23 Monoclonal Antibody for Western Blot and ELISA
  • Host Rat
  • Target Species Mouse
  • Conjugate Unconjugated
  • Catalog # MA523793
  • Size 500 µg
Applications
ELISA (0.1-0.4 µg/ml (Detection), 2-8 µg/ml (Capture)), Western Blot (1 µg/ml)
Alternate names:
Interleukin-23 subunit p19
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA523793 349.00
Check your price
  • IL-12/IL-23 p40 Polyclonal Antibody for Western Blot and Neu
  • Host Goat
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # PA547003
  • Size 100 µg
Applications
Neutralization (0.004-0.02 µg/ml), Western Blot (1 µg/ml)
Alternate names:
NK cell stimulatory factor chain 2; IL-12 subunit p40; Cytotoxic lymphocyte maturation factor 40 kDa subunit
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA547003 375.00
Check your price
  • IL-12/IL-23 p40 Monoclonal Antibody, PE conjugate for Flow
  • Host Rat
  • Target Species Mouse
  • Conjugate PE
  • Catalog # A18694
  • Size 25 µg
Applications
Flow Cytometry (Assay Dependent)
Alternate names:
IL-12 subunit p40; Cytotoxic lymphocyte maturation factor 40 kDa subunit
Catalog # Size Price (AUD) Your Price (AUD) Qty
A18694 117.00
Check your price
  • IL-12/IL-23 p40 Monoclonal Antibody for ELISA
  • Host Mouse
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # MA523724
  • Size 500 µg
Applications
ELISA (0.1-0.4 µg/ml (Detection), 2-8 µg/ml (Capture))
Alternate names:
NK cell stimulatory factor chain 2; IL-12 subunit p40; Cytotoxic lymphocyte maturation factor 40 kDa subunit
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA523724 349.00
Check your price
  • IL-12/IL-23 p40 Monoclonal Antibody for ELISA
  • Host Rat
  • Target Species Mouse
  • Conjugate Unconjugated
  • Catalog # MA523723
  • Size 500 µg
Applications
ELISA (0.1-0.4 µg/ml (Detection), 2-8 µg/ml (Capture))
Alternate names:
IL-12 subunit p40; Cytotoxic lymphocyte maturation factor 40 kDa subunit
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA523723 349.00
Check your price
  • IL-12/IL-23 p40 Polyclonal Antibody for Western Blot and Neu
  • Host Goat
  • Target Species Dog
  • Conjugate Unconjugated
  • Catalog # PA546917
  • Size 100 µg
Applications
Neutralization (0.1-0.4 µg/ml), Western Blot (0.1 µg/ml)
Alternate names:
IL-12 subunit p40; Cytotoxic lymphocyte maturation factor 40 kDa subunit
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA546917 375.00
Check your price
  • IL23R Monoclonal Antibody for Western Blot and Neu
  • Host Rat
  • Target Species Mouse
  • Conjugate Unconjugated
  • Catalog # MA523906
  • Size 100 µg
Applications
Neutralization (0.1-0.4 µg/ml), Western Blot (1 µg/ml)
Alternate names:
IBD17, IL-23 receptor, IL-23R, interleukin 23 receptor, interleukin-23 receptor
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA523906 275.00
Check your price
  • IL23R Polyclonal Antibody for Western Blot and Neu
  • Host Goat
  • Target Species Mouse
  • Conjugate Unconjugated
  • Catalog # PA547184
  • Size 100 µg
Applications
Blocking Assay (Assay-Dependent), Western Blot (0.1 µg/ml)
Alternate names:
IBD17, IL-23 receptor, IL-23R, interleukin 23 receptor, interleukin-23 receptor
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA547184 349.00
Check your price
  • IL-12/IL-23 p40 Monoclonal Antibody, APC conjugate for Flow
  • Host Rat
  • Target Species Mouse
  • Conjugate APC
  • Catalog # MA523652
  • Size 100 Tests
Applications
Flow Cytometry (Assay-Dependent)
Alternate names:
IL-12 subunit p40; Cytotoxic lymphocyte maturation factor 40 kDa subunit
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA523652 339.00
Check your price
  • IL-22 Oligoclonal Antibody for Western Blot
  • Host Rabbit
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # 710084
  • Size 100 µg
Applications
Western Blot (1-5 ug/ml)
Alternate names:
IL-10-related T-cell-derived-inducible factor; Cytokine Zcyto18
Catalog # Size Price (AUD) Your Price (AUD) Qty
710084 309.00
Check your price
  • IL-22 Monoclonal Antibody for Western Blot
  • Host Rabbit
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # 701031
  • Size 100 µg
Applications
Western Blot (4-5 ug/ml)
Alternate names:
IL-10-related T-cell-derived-inducible factor; Cytokine Zcyto18
Catalog # Size Price (AUD) Your Price (AUD) Qty
701031 309.00
Check your price
  • IL-12/IL-23 p40 Monoclonal Antibody for ELISA
  • Host Mouse
  • Target Species Pig
  • Conjugate Unconjugated
  • Catalog # MA523768
  • Size 500 µg
Applications
ELISA (0.5-2 µg/ml (Detection), 2-8 µg/ml (Capture))
Alternate names:
IL-12 subunit p40; Cytotoxic lymphocyte maturation factor 40 kDa subunit
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA523768 349.00
Check your price
  • IL23R Monoclonal Antibody for Western Blot
  • Host Mouse
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # MA523935
  • Size 100 µg
Applications
Western Blot (2 µg/ml)
Alternate names:
IBD17, IL-23 receptor, IL-23R, interleukin 23 receptor, interleukin-23 receptor
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA523935 275.00
Check your price
  • IL-12 p40 Polyclonal Antibody for Western Blot
  • Host Goat
  • Target Species Mouse, Rat
  • Conjugate Unconjugated
  • Catalog # PA518741
  • Size 100 µg
Applications
Western Blot (1-3 ug/ml)
Alternate names:
IL-12 subunit p40; Cytotoxic lymphocyte maturation factor 40 kDa subunit
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA518741 349.00
Check your price
  • IL-12 p70 Monoclonal Antibody for Western Blot and ELISA
  • Host Rat
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # M122
  • Size 500 µg
Applications
ELISA (Assay Dependent), Western Blot (Assay Dependent)
Alternate names:
NK cell stimulatory factor chain 1; IL-12 subunit p35; Cytotoxic lymphocyte maturation factor 35 kDa subunit; NK cell stimulatory factor chain 2; IL-12 subunit p40; Cytotoxic lymphocyte maturation factor 40 kDa subunit
Catalog # Size Price (AUD) Your Price (AUD) Qty
M122 375.00
Check your price
  • IL-27 Polyclonal Antibody for Western Blot
  • Host Rabbit
  • Target Species Human, Mouse
  • Conjugate Unconjugated
  • Catalog # PA520241
  • Size 100 ug
Applications
Western Blot (2-4 ug/ml)
Alternate names:
Epstein-Barr virus-induced gene 3 protein homolog; p28; Interleukin-30; Epstein-Barr virus-induced gene 3 protein
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA520241 349.00
Check your price
  • IL-27 Polyclonal Antibody for Western Blot and ICC
  • Host Rabbit
  • Target Species Human, Mouse
  • Conjugate Unconjugated
  • Catalog # PA520240
  • Size 100 ug
Applications
Immunocytochemistry (10 µg/ml), Western Blot (2-4 ug/ml)
Alternate names:
Epstein-Barr virus-induced gene 3 protein homolog; p28; Interleukin-30; Epstein-Barr virus-induced gene 3 protein
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA520240 349.00
Check your price
  • IL-17A Monoclonal Antibody for IP, ELISA and Neu
  • Host Mouse
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # MA523738
  • Size 500 µg
Applications
ELISA (0.1-0.4 µg/ml (Detection), 2-8 µg/ml (Capture)), Immunoprecipitation (3 µg/100 µg cell lysate), Neutralization (1-3 µg/ml)
Alternate names:
Cytotoxic T-lymphocyte-associated antigen 8
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA523738 349.00
Check your price
  • IL-17A Monoclonal Antibody, Biotin conjugate for ELISA
  • Host Mouse
  • Target Species Human
  • Conjugate Biotin
  • Catalog # MA1069B
  • Size 100 µg
Applications
ELISA (0.125-0.25 µg/ml)
Alternate names:
Cytotoxic T-lymphocyte-associated antigen 8
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA1069B 349.00
Check your price
  • IL-12 p70 Monoclonal Antibody for IF, ICC and ELISA
  • Host Mouse
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # MA523715
  • Size 500 µg
Applications
ELISA (0.1-0.4 µg/ml (Detection), 2-8 µg/ml (Capture)), Immunocytochemistry (8-25 µg/ml), Immunofluorescence (8-25 µg/ml)
Alternate names:
NK cell stimulatory factor chain 1; IL-12 subunit p35; Cytotoxic lymphocyte maturation factor 35 kDa subunit; NK cell stimulatory factor chain 2; IL-12 subunit p40; Cytotoxic lymphocyte maturation factor 40 kDa subunit
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA523715 349.00
Check your price
  • IL-12 p40 Oligoclonal Antibody for Western Blot
  • Host Rabbit
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # 710257
  • Size 100 µg
Applications
Western Blot (1-3 ug/ml)
Alternate names:
NK cell stimulatory factor chain 2; IL-12 subunit p40; Cytotoxic lymphocyte maturation factor 40 kDa subunit
Catalog # Size Price (AUD) Your Price (AUD) Qty
710257 309.00
Check your price
  • IL-12 p40 Monoclonal Antibody for Western Blot
  • Host Rabbit
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # 701233
  • Size 100 µg
Applications
Western Blot (0.5-2 ug/ml)
Alternate names:
NK cell stimulatory factor chain 2; IL-12 subunit p40; Cytotoxic lymphocyte maturation factor 40 kDa subunit
Catalog # Size Price (AUD) Your Price (AUD) Qty
701233 309.00
Check your price