1 - 15 of 15 products displayed
  • IFNLR1 Monoclonal Antibody for Flow
  • Host Mouse
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # MA524271
  • Size 100µg
Applications
Flow Cytometry (0.25 µg/10^6 cells)
Alternate names:
Likely interleukin or cytokine receptor 2; Interleukin-28 receptor subunit alpha; Cytokine receptor family 2 member 12; Cytokine receptor class-II member 12
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA524271 275.00
Check your price
  • IFNLR1 Polyclonal Antibody for Western Blot and Flow
  • Host Sheep
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # PA547713
  • Size 100µg
Applications
Flow Cytometry (2.5 µg/10^6 cells), Western Blot (2 µg/ml)
Alternate names:
Likely interleukin or cytokine receptor 2; Interleukin-28 receptor subunit alpha; Cytokine receptor family 2 member 12; Cytokine receptor class-II member 12
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA547713 349.00
Check your price
  • IFNLR1 Polyclonal Antibody for IF, ICC, IHC (P) and ELISA
  • Host Goat
  • Target Species Mouse, Rat
  • Conjugate Unconjugated
  • Catalog # PA121360
  • Size 100µg
Applications
ELISA (1:100,000), Immunocytochemistry (1:300), Immunofluorescence (1:300), Immunohistochemistry (Paraffin) (Assay-dependent)
Alternate names:
Interleukin-28 receptor subunit alpha; Cytokine receptor family 2 member 12; Cytokine receptor class-II member 12
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA121360 425.00
Check your price
  • IL-29/IL-28B Monoclonal Antibody for ELISA and Neu
  • Host Mouse
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # MA523791
  • Size 500µg
Applications
ELISA (0.1-0.4 µg/ml (Detection), 2-8 µg/ml (Capture)), Neutralization (1-4 µg/ml)
Alternate names:
Interleukin-28C; Interleukin-28B; Cytokine Zcyto22; Interleukin-29; Cytokine Zcyto21
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA523791 349.00
Check your price
  • IL-28A Polyclonal Antibody for Western Blot, IF, ICC and Neu
  • Host Goat
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # PA547163
  • Size 100µg
Applications
Immunocytochemistry (5-15 µg/ml), Immunofluorescence (5-15 µg/ml), Neutralization (0.3-1.5 µg/ml), Western Blot (0.1 µg/ml)
Alternate names:
Interleukin-28A; Cytokine Zcyto20
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA547163 349.00
Check your price
  • IL-29 Polyclonal Antibody for Western Blot and Neu
  • Host Goat
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # PA547165
  • Size 100µg
Applications
Neutralization (0.2-1.0 µg/ml), Western Blot (0.1 µg/ml)
Alternate names:
Interleukin-29; Cytokine Zcyto21
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA547165 349.00
Check your price
  • IL-27 Polyclonal Antibody for Western Blot, ELISA and Neu
  • Host Goat
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # PA546993
  • Size 100µg
Applications
ELISA (0.1-0.4 µg/ml (Detection), 0.2-0.8 µg/ml (Capture)), Neutralization (1-5 µg/ml), Western Blot (1 µg/ml)
Alternate names:
p28; Interleukin-30; Epstein-Barr virus-induced gene 3 protein
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA546993 375.00
Check your price
  • IL-28B Monoclonal Antibody for Western Blot
  • Host Mouse
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # MA524254
  • Size 100µg
Applications
Western Blot (2 µg/ml)
Alternate names:
Interleukin-28C; Interleukin-28B; Cytokine Zcyto22
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA524254 275.00
Check your price
  • IL-29 Monoclonal Antibody for ELISA and Neu
  • Host Mouse
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # MA523903
  • Size 100µg
Applications
ELISA (0.1-0.4 µg/ml (Detection), 2-8 µg/ml (Capture)), Neutralization (1-4 µg/ml)
Alternate names:
Interleukin-29; Cytokine Zcyto21
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA523903 275.00
Check your price
  • IL-28A/IL-28B Monoclonal Antibody for ELISA and Neu
  • Host Rat
  • Target Species Mouse
  • Conjugate Unconjugated
  • Catalog # MA523902
  • Size 100µg
Applications
ELISA (0.5-2 µg/ml (Detection), 2-8 µg/ml (Capture)), Neutralization (3-9 µg/ml)
Alternate names:
Interleukin-28B; Interleukin-28A
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA523902 275.00
Check your price
  • IL-28B Polyclonal Antibody for Western Blot and Neu
  • Host Goat
  • Target Species Mouse
  • Conjugate Unconjugated
  • Catalog # PA547202
  • Size 100µg
Applications
Neutralization (2-10 µg/ml), Western Blot (0.1 µg/ml)
Alternate names:
Interleukin-28B
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA547202 349.00
Check your price
  • CD25 Monoclonal Antibody for Western Blot, IF, ICC, IHC, Flow and IP
  • Host Mouse
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # M10AR
  • Size 100tests
Applications
Flow Cytometry (Assay Dependent), Immunocytochemistry (1:10-1:100), Immunofluorescence (1:10-1:100), Immunohistochemistry (Assay Dependent), Immunoprecipitation (Assay Dependent), Western Blot (Assay Dependent)
Alternate names:
p55; TAC antigen
Catalog # Size Price (AUD) Your Price (AUD) Qty
M10AR 275.00
Check your price
  • IL-29 Polyclonal Antibody for Western Blot and IHC (P)
  • Host Rabbit
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # PA511710
  • Size 400µL
Applications
Immunohistochemistry (1:50-1:100), Western Blot (1:1000)
Alternate names:
Interleukin-29; Cytokine Zcyto21
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA511710 349.00
Check your price
  • IL-28A Polyclonal Antibody for Western Blot
  • Host Rabbit
  • Target Species Human, Mouse
  • Conjugate Unconjugated
  • Catalog # PA511708
  • Size 400µL
Applications
Western Blot (1:1000)
Alternate names:
Interleukin-28A; Cytokine Zcyto20
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA511708 349.00
Check your price
  • CD81 Monoclonal Antibody for Western Blot, IHC (F), Flow, IP and ELISA
  • Host Hamster
  • Target Species Mouse, Rat
  • Conjugate Unconjugated
  • Catalog # MA180209
  • Size 100µg
Applications
ELISA (Assay Dependent), Flow Cytometry (1/10-1/50), Immunohistochemistry (Frozen) (Assay Dependent), Immunoprecipitation (Assay Dependent), Western Blot (Assay Dependent)
Alternate names:
Target of the antiproliferative antibody 1; 26 kDa cell surface protein TAPA-1
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA180209 159.00
Check your price